Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LASSWPPLEL QSSNQSQLFP QNATACDNAP EAWDLLHRVL PTFIISICFF GLLGNLFVLL VFLLPRRQLN VAEIYLANLA ASDLVFVLGL PFWAENIWNQ FNWPFGALLC RVINGVIKAN LFISIFLVVA ISQDRYRVLV HPMASRRQQR RRQARVTCVL IWVVGGLLSI PTFLLRSIQA VPDLNITACI LLLPHEAWHF ARIVELNILG FLLPLAAIVF FNYHILASLR TREEVSRTRC GGRKDSKTTA LILTLVVAFL VCWAPYHFFA FLEFLFQVQA VRGCFWEDFI DLGLQLANFF AFTNSSLNPV IYVFVGRLFR TKVWELYKQC TPKSLAPISS SHRKEIFQLF WRN
Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B1 bradykinin receptor; B1R; Bk b1 receptor; BK-1 receptor; BKRB1; bradykinin B1 receptor; bradykinin receptor 1; bradykinin receptor B2
Gene Aliases: B1BKR; B1R; BDKRB1; BDKRB2; BKB1R; BKR1; BRADYB1
UniProt ID: (Human) P46663
Entrez Gene ID: (Human) 623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support