Product References
Effects of a High-Protein Diet on Kidney Injury under Conditions of Non-CKD or CKD in Mice.
International journal of molecular sciences
Tanaka S,Wakui H,Azushima K,Tsukamoto S,Yamaji T,Urate S,Suzuki T,Abe E,Taguchi S,Yamada T,Kobayashi R,Kanaoka T,Kamimura D,Kinguchi S,Takiguchi M,Funakoshi K,Yamashita A,Ishigami T,Tamura K
Mon Apr 24 00:00:00 EDT 2023
Cryopreservation of Fetal Porcine Kidneys for Xenogeneic Regenerative Medicine.
Journal of clinical medicine
Matsui K,Kinoshita Y,Inage Y,Matsumoto N,Morimoto K,Saito Y,Takamura T,Matsunari H,Yamanaka S,Nagashima H,Kobayashi E,Yokoo T
Wed Mar 15 00:00:00 EDT 2023
DACH1 regulates macrophage activation and tumour progression in hypopharyngeal squamous cell carcinoma.
Immunology
Li W,Xu L,Cao J,Ge J,Liu X,Liu P,Teng Y,Wang S,Sun Y,Liu M,Tian L
Sun Oct 01 00:00:00 EDT 2023
DACH1 regulates macrophage activation and tumour progression in hypopharyngeal squamous cell carcinoma.
Immunology
Li W,Xu L,Cao J,Ge J,Liu X,Liu P,Teng Y,Wang S,Sun Y,Liu M,Tian L
Sun Oct 01 00:00:00 EDT 2023
DACH1 regulates macrophage activation and tumour progression in hypopharyngeal squamous cell carcinoma.
Immunology
Li W,Xu L,Cao J,Ge J,Liu X,Liu P,Teng Y,Wang S,Sun Y,Liu M,Tian L
Sun Oct 01 00:00:00 EDT 2023
Dach1 deficiency drives alveolar epithelium apoptosis in pulmonary fibrosis via modulating C-Jun/Bim activity.
Translational research : the journal of laboratory and clinical medicine
Lu Y,Tang K,Wang S,Tian Z,Fan Y,Li B,Wang M,Zhao J,Xie J
Sat Jul 01 00:00:00 EDT 2023
Dach1 deficiency drives alveolar epithelium apoptosis in pulmonary fibrosis via modulating C-Jun/Bim activity.
Translational research : the journal of laboratory and clinical medicine
Lu Y,Tang K,Wang S,Tian Z,Fan Y,Li B,Wang M,Zhao J,Xie J
Sat Jul 01 00:00:00 EDT 2023
Dach1 deficiency drives alveolar epithelium apoptosis in pulmonary fibrosis via modulating C-Jun/Bim activity.
Translational research : the journal of laboratory and clinical medicine
Lu Y,Tang K,Wang S,Tian Z,Fan Y,Li B,Wang M,Zhao J,Xie J
Sat Jul 01 00:00:00 EDT 2023
Dach1 deficiency drives alveolar epithelium apoptosis in pulmonary fibrosis via modulating C-Jun/Bim activity.
Translational research : the journal of laboratory and clinical medicine
Lu Y,Tang K,Wang S,Tian Z,Fan Y,Li B,Wang M,Zhao J,Xie J
Sat Jul 01 00:00:00 EDT 2023
An integrated co-expression network analysis reveals novel genetic biomarkers for immune cell infiltration in chronic kidney disease.
Frontiers in immunology
Xia J,Hou Y,Cai A,Xu Y,Yang W,Huang M,Mou S
Tue Mar 07 00:00:00 EST 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Research square
Li Z,Jiao X,Robertson AG,Sante GD,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJ,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Tue Jan 10 00:00:00 EST 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Research square
Li Z,Jiao X,Robertson AG,Sante GD,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJ,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Tue Jan 10 00:00:00 EST 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Research square
Li Z,Jiao X,Robertson AG,Sante GD,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJ,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Tue Jan 10 00:00:00 EST 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Research square
Li Z,Jiao X,Robertson AG,Sante GD,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJ,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Tue Jan 10 00:00:00 EST 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Oncogene
Li Z,Jiao X,Robertson AG,Di Sante G,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJM,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Thu Jun 01 00:00:00 EDT 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Oncogene
Li Z,Jiao X,Robertson AG,Di Sante G,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJM,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Thu Jun 01 00:00:00 EDT 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Oncogene
Li Z,Jiao X,Robertson AG,Di Sante G,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJM,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Thu Jun 01 00:00:00 EDT 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Oncogene
Li Z,Jiao X,Robertson AG,Di Sante G,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJM,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Thu Jun 01 00:00:00 EDT 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Oncogene
Li Z,Jiao X,Robertson AG,Di Sante G,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJM,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Thu Jun 01 00:00:00 EDT 2023
The DACH1 gene is frequently deleted in prostate cancer, restrains prostatic intraepithelial neoplasia, decreases DNA damage repair, and predicts therapy responses.
Oncogene
Li Z,Jiao X,Robertson AG,Di Sante G,Ashton AW,DiRocco A,Wang M,Zhao J,Addya S,Wang C,McCue PA,South AP,Cordon-Cardo C,Liu R,Patel K,Hamid R,Parmar J,DuHadaway JB,Jones SJM,Casimiro MC,Schultz N,Kossenkov A,Phoon LY,Chen H,Lan L,Sun Y,Iczkowski KA,Rui H,Pestell RG
Thu Jun 01 00:00:00 EDT 2023
Derivation of Multipotent Neural Progenitors from Human Embryonic Stem Cells for Cell Therapy and Biomedical Applications.
Methods in molecular biology (Clifton, N.J.)
Vitillo L,Vallier L
Wed Jul 27 00:00:00 EDT 2022
Haploinsufficiency of the mouse Tshz3 gene leads to kidney defects.
Human molecular genetics
Sanchez-Martin I,Magalhães P,Ranjzad P,Fatmi A,Richard F,Manh TPV,Saurin AJ,Feuillet G,Denis C,Woolf AS,Schanstra JP,Zürbig P,Caubit X,Fasano L
Wed Jun 22 00:00:00 EDT 2022
Establishment of autaptic culture with human-induced pluripotent stem cell-derived astrocytes.
iScience
Uchino K,Tanaka Y,Kawaguchi S,Kubota K,Watanabe T,Katsurabayashi S,Hirose S,Iwasaki K
Fri Aug 19 00:00:00 EDT 2022
Diastolic dysfunction in a pre-clinical model of diabetes is associated with changes in the cardiac non-myocyte cellular composition.
Cardiovascular diabetology
Cohen CD,De Blasio MJ,Lee MKS,Farrugia GE,Prakoso D,Krstevski C,Deo M,Donner DG,Kiriazis H,Flynn MC,Gaynor TL,Murphy AJ,Drummond GR,Pinto AR,Ritchie RH
Tue Jun 01 00:00:00 EDT 2021
FOXM1-induced miR-552 expression contributes to pancreatic cancer progression by targeting multiple tumor suppressor genes.
International journal of biological sciences
Wang X,Dou N,Wang J,Zhang Y,Li Y,Gao Y
Fri Jan 21 00:00:00 EST 2022
Generation of Human iPSC-derived Neural Progenitor Cells (NPCs) as Drug Discovery Model for Neurological and Mitochondrial Disorders.
Bio-protocol
Zink A,Lisowski P,Prigione A
Fri Mar 05 00:00:00 EST 2021
DACH1 protects podocytes from experimental diabetic injury and modulates PTIP-H3K4Me3 activity.
The Journal of clinical investigation
Cao A,Li J,Asadi M,Basgen JM,Zhu B,Yi Z,Jiang S,Doke T,El Shamy O,Patel N,Cravedi P,Azeloglu EU,Campbell KN,Menon M,Coca S,Zhang W,Wang H,Zen K,Liu Z,Murphy B,He JC,D'Agati VD,Susztak K,Kaufman L
Mon May 17 00:00:00 EDT 2021
Podometrics in Japanese Living Donor Kidneys: Associations with Nephron Number, Age, and Hypertension.
Journal of the American Society of Nephrology : JASN
Haruhara K,Sasaki T,de Zoysa N,Okabayashi Y,Kanzaki G,Yamamoto I,Harper IS,Puelles VG,Shimizu A,Cullen-McEwen LA,Tsuboi N,Yokoo T,Bertram JF
Mon May 03 00:00:00 EDT 2021
Organoid modelling identifies that DACH1 functions as a tumour promoter in colorectal cancer by modulating BMP signalling.
EBioMedicine
Hu X,Zhang L,Li Y,Ma X,Dai W,Gao X,Rao X,Fu G,Wang R,Pan M,Guo Q,Xu X,Zhou Y,Gao J,Zhang Z,Cai S,Peng J,Hua G
Mon Jun 01 00:00:00 EDT 2020
Organoid modelling identifies that DACH1 functions as a tumour promoter in colorectal cancer by modulating BMP signalling.
EBioMedicine
Hu X,Zhang L,Li Y,Ma X,Dai W,Gao X,Rao X,Fu G,Wang R,Pan M,Guo Q,Xu X,Zhou Y,Gao J,Zhang Z,Cai S,Peng J,Hua G
Mon Jun 01 00:00:00 EDT 2020
Organoid modelling identifies that DACH1 functions as a tumour promoter in colorectal cancer by modulating BMP signalling.
EBioMedicine
Hu X,Zhang L,Li Y,Ma X,Dai W,Gao X,Rao X,Fu G,Wang R,Pan M,Guo Q,Xu X,Zhou Y,Gao J,Zhang Z,Cai S,Peng J,Hua G
Mon Jun 01 00:00:00 EDT 2020
Global polysome analysis of normal and injured podocytes.
American journal of physiology. Renal physiology
Okabe M,Motojima M,Miyazaki Y,Pastan I,Yokoo T,Matsusaka T
Fri Feb 01 00:00:00 EST 2019
Global polysome analysis of normal and injured podocytes.
American journal of physiology. Renal physiology
Okabe M,Motojima M,Miyazaki Y,Pastan I,Yokoo T,Matsusaka T
Fri Feb 01 00:00:00 EST 2019
Global polysome analysis of normal and injured podocytes.
American journal of physiology. Renal physiology
Okabe M,Motojima M,Miyazaki Y,Pastan I,Yokoo T,Matsusaka T
Fri Feb 01 00:00:00 EST 2019
MicroRNA-6807-3p promotes the tumorigenesis of glioma by targeting downstream DACH1.
Brain research
Lu GF,Geng F,Xiao Z,Chen YS,Han Y,You CY,Gong NL,Xie ZM,Pan M
Mon Apr 01 00:00:00 EDT 2019
MicroRNA-6807-3p promotes the tumorigenesis of glioma by targeting downstream DACH1.
Brain research
Lu GF,Geng F,Xiao Z,Chen YS,Han Y,You CY,Gong NL,Xie ZM,Pan M
Mon Apr 01 00:00:00 EDT 2019
MicroRNA-6807-3p promotes the tumorigenesis of glioma by targeting downstream DACH1.
Brain research
Lu GF,Geng F,Xiao Z,Chen YS,Han Y,You CY,Gong NL,Xie ZM,Pan M
Mon Apr 01 00:00:00 EDT 2019
MicroRNA-6807-3p promotes the tumorigenesis of glioma by targeting downstream DACH1.
Brain research
Lu GF,Geng F,Xiao Z,Chen YS,Han Y,You CY,Gong NL,Xie ZM,Pan M
Mon Apr 01 00:00:00 EDT 2019
CXCL1 as an Unfavorable Prognosis Factor Negatively Regulated by DACH1 in Non-small Cell Lung Cancer.
Frontiers in oncology
Yu S,Yi M,Xu L,Qin S,Li A,Wu K
Tue Sep 29 00:00:00 EDT 2020
Dachshund Depletion Disrupts Mammary Gland Development and Diverts the Composition of the Mammary Gland Progenitor Pool.
Stem cell reports
Jiao X,Li Z,Wang M,Katiyar S,Di Sante G,Farshchian M,South AP,Cocola C,Colombo D,Reinbold R,Zucchi I,Wu K,Tabas I,Spike BT,Pestell RG
Tue Jan 08 00:00:00 EST 2019
Dachshund Depletion Disrupts Mammary Gland Development and Diverts the Composition of the Mammary Gland Progenitor Pool.
Stem cell reports
Jiao X,Li Z,Wang M,Katiyar S,Di Sante G,Farshchian M,South AP,Cocola C,Colombo D,Reinbold R,Zucchi I,Wu K,Tabas I,Spike BT,Pestell RG
Tue Jan 08 00:00:00 EST 2019
Dachshund Depletion Disrupts Mammary Gland Development and Diverts the Composition of the Mammary Gland Progenitor Pool.
Stem cell reports
Jiao X,Li Z,Wang M,Katiyar S,Di Sante G,Farshchian M,South AP,Cocola C,Colombo D,Reinbold R,Zucchi I,Wu K,Tabas I,Spike BT,Pestell RG
Tue Jan 08 00:00:00 EST 2019
Dachshund Depletion Disrupts Mammary Gland Development and Diverts the Composition of the Mammary Gland Progenitor Pool.
Stem cell reports
Jiao X,Li Z,Wang M,Katiyar S,Di Sante G,Farshchian M,South AP,Cocola C,Colombo D,Reinbold R,Zucchi I,Wu K,Tabas I,Spike BT,Pestell RG
Tue Jan 08 00:00:00 EST 2019
Expression of transcription factors and signaling molecules in the cerebellar granule cell development.
Gene expression patterns : GEP
Shiraishi RD,Miyashita S,Yamashita M,Adachi T,Shimoda MM,Owa T,Hoshino M
Sun Dec 01 00:00:00 EST 2019
Expression of transcription factors and signaling molecules in the cerebellar granule cell development.
Gene expression patterns : GEP
Shiraishi RD,Miyashita S,Yamashita M,Adachi T,Shimoda MM,Owa T,Hoshino M
Sun Dec 01 00:00:00 EST 2019
A single-cell transcriptional roadmap for cardiopharyngeal fate diversification.
Nature cell biology
Wang W,Niu X,Stuart T,Jullian E,Mauck WM,Kelly RG,Satija R,Christiaen L
Sat Jun 01 00:00:00 EDT 2019
Single-cell analysis of early progenitor cells that build coronary arteries.
Nature
Su T,Stanley G,Sinha R,D'Amato G,Das S,Rhee S,Chang AH,Poduri A,Raftrey B,Dinh TT,Roper WA,Li G,Quinn KE,Caron KM,Wu S,Miquerol L,Butcher EC,Weissman I,Quake S,Red-Horse K
Sun Jul 01 00:00:00 EDT 2018
Single-cell analysis of early progenitor cells that build coronary arteries.
Nature
Su T,Stanley G,Sinha R,D'Amato G,Das S,Rhee S,Chang AH,Poduri A,Raftrey B,Dinh TT,Roper WA,Li G,Quinn KE,Caron KM,Wu S,Miquerol L,Butcher EC,Weissman I,Quake S,Red-Horse K
Sun Jul 01 00:00:00 EDT 2018
Genome Editing in Neuroepithelial Stem Cells to Generate Human Neurons with High Adenosine-Releasing Capacity.
Stem cells translational medicine
Poppe D,Doerr J,Schneider M,Wilkens R,Steinbeck JA,Ladewig J,Tam A,Paschon DE,Gregory PD,Reik A,Müller CE,Koch P,Brüstle O
Fri Jun 01 00:00:00 EDT 2018
Genome Editing in Neuroepithelial Stem Cells to Generate Human Neurons with High Adenosine-Releasing Capacity.
Stem cells translational medicine
Poppe D,Doerr J,Schneider M,Wilkens R,Steinbeck JA,Ladewig J,Tam A,Paschon DE,Gregory PD,Reik A,Müller CE,Koch P,Brüstle O
Fri Jun 01 00:00:00 EDT 2018
Modulation of inflammatory and hormonal parameters in response to testosterone therapy: Effects on the ventral prostate of adult rats.
Cell biology international
Mendes LO,Castilho ACS,Pinho CF,Gonçalvez BF,Razza EM,Chuffa LGA,Anselmo-Franci JA,Scarano WR,Martinez FE
Sat Sep 01 00:00:00 EDT 2018
Modulation of inflammatory and hormonal parameters in response to testosterone therapy: Effects on the ventral prostate of adult rats.
Cell biology international
Mendes LO,Castilho ACS,Pinho CF,Gonçalvez BF,Razza EM,Chuffa LGA,Anselmo-Franci JA,Scarano WR,Martinez FE
Sat Sep 01 00:00:00 EDT 2018
Dachshund 1 is Differentially Expressed Between Male and Female Breast Cancer: A Matched Case-Control Study of Clinical Characteristics and Prognosis.
Clinical breast cancer
Cui Q,Kong D,Li Z,Ahiable P,Wang K,Wu K,Wu G
Mon Oct 01 00:00:00 EDT 2018
Dachshund 1 is Differentially Expressed Between Male and Female Breast Cancer: A Matched Case-Control Study of Clinical Characteristics and Prognosis.
Clinical breast cancer
Cui Q,Kong D,Li Z,Ahiable P,Wang K,Wu K,Wu G
Mon Oct 01 00:00:00 EDT 2018
CCBE1 is required for coronary vessel development and proper coronary artery stem formation in the mouse heart.
Developmental dynamics : an official publication of the American Association of Anatomists
Bonet F,Pereira PNG,Bover O,Marques S,Inácio JM,Belo JA
Mon Oct 01 00:00:00 EDT 2018
CCBE1 is required for coronary vessel development and proper coronary artery stem formation in the mouse heart.
Developmental dynamics : an official publication of the American Association of Anatomists
Bonet F,Pereira PNG,Bover O,Marques S,Inácio JM,Belo JA
Mon Oct 01 00:00:00 EDT 2018
Endothelial deletion of Ino80 disrupts coronary angiogenesis and causes congenital heart disease.
Nature communications
Rhee S,Chung JI,King DA,D'amato G,Paik DT,Duan A,Chang A,Nagelberg D,Sharma B,Jeong Y,Diehn M,Wu JC,Morrison AJ,Red-Horse K
Thu Jan 25 00:00:00 EST 2018
Endothelial deletion of Ino80 disrupts coronary angiogenesis and causes congenital heart disease.
Nature communications
Rhee S,Chung JI,King DA,D'amato G,Paik DT,Duan A,Chang A,Nagelberg D,Sharma B,Jeong Y,Diehn M,Wu JC,Morrison AJ,Red-Horse K
Thu Jan 25 00:00:00 EST 2018
Metformin blocks myeloid-derived suppressor cell accumulation through AMPK-DACH1-CXCL1 axis.
Oncoimmunology
Qin G,Lian J,Huang L,Zhao Q,Liu S,Zhang Z,Chen X,Yue D,Li L,Li F,Wang L,Umansky V,Zhang B,Yang S,Zhang Y
Tue Jan 12 00:00:00 EST 2021
Metformin blocks myeloid-derived suppressor cell accumulation through AMPK-DACH1-CXCL1 axis.
Oncoimmunology
Qin G,Lian J,Huang L,Zhao Q,Liu S,Zhang Z,Chen X,Yue D,Li L,Li F,Wang L,Umansky V,Zhang B,Yang S,Zhang Y
Tue Jan 12 00:00:00 EST 2021
Metformin blocks myeloid-derived suppressor cell accumulation through AMPK-DACH1-CXCL1 axis.
Oncoimmunology
Qin G,Lian J,Huang L,Zhao Q,Liu S,Zhang Z,Chen X,Yue D,Li L,Li F,Wang L,Umansky V,Zhang B,Yang S,Zhang Y
Tue Jan 12 00:00:00 EST 2021
Metformin blocks myeloid-derived suppressor cell accumulation through AMPK-DACH1-CXCL1 axis.
Oncoimmunology
Qin G,Lian J,Huang L,Zhao Q,Liu S,Zhang Z,Chen X,Yue D,Li L,Li F,Wang L,Umansky V,Zhang B,Yang S,Zhang Y
Tue Jan 12 00:00:00 EST 2021
Metformin blocks myeloid-derived suppressor cell accumulation through AMPK-DACH1-CXCL1 axis.
Oncoimmunology
Qin G,Lian J,Huang L,Zhao Q,Liu S,Zhang Z,Chen X,Yue D,Li L,Li F,Wang L,Umansky V,Zhang B,Yang S,Zhang Y
Tue Jan 12 00:00:00 EST 2021
Metformin blocks myeloid-derived suppressor cell accumulation through AMPK-DACH1-CXCL1 axis.
Oncoimmunology
Qin G,Lian J,Huang L,Zhao Q,Liu S,Zhang Z,Chen X,Yue D,Li L,Li F,Wang L,Umansky V,Zhang B,Yang S,Zhang Y
Tue Jan 12 00:00:00 EST 2021
DACH1 antagonizes CXCL8 to repress tumorigenesis of lung adenocarcinoma and improve prognosis.
Journal of hematology & oncology
Liu Q,Li A,Yu S,Qin S,Han N,Pestell RG,Han X,Wu K
Tue Apr 10 00:00:00 EDT 2018
DACH1 antagonizes CXCL8 to repress tumorigenesis of lung adenocarcinoma and improve prognosis.
Journal of hematology & oncology
Liu Q,Li A,Yu S,Qin S,Han N,Pestell RG,Han X,Wu K
Tue Apr 10 00:00:00 EDT 2018
DACH1 antagonizes CXCL8 to repress tumorigenesis of lung adenocarcinoma and improve prognosis.
Journal of hematology & oncology
Liu Q,Li A,Yu S,Qin S,Han N,Pestell RG,Han X,Wu K
Tue Apr 10 00:00:00 EDT 2018
DACH1 antagonizes CXCL8 to repress tumorigenesis of lung adenocarcinoma and improve prognosis.
Journal of hematology & oncology
Liu Q,Li A,Yu S,Qin S,Han N,Pestell RG,Han X,Wu K
Tue Apr 10 00:00:00 EDT 2018
Breathing control center neurons that promote arousal in mice.
Science (New York, N.Y.)
Yackle K,Schwarz LA,Kam K,Sorokin JM,Huguenard JR,Feldman JL,Luo L,Krasnow MA
Fri Mar 31 00:00:00 EDT 2017
Breathing control center neurons that promote arousal in mice.
Science (New York, N.Y.)
Yackle K,Schwarz LA,Kam K,Sorokin JM,Huguenard JR,Feldman JL,Luo L,Krasnow MA
Fri Mar 31 00:00:00 EDT 2017
CAMKIIγ suppresses an efferocytosis pathway in macrophages and promotes atherosclerotic plaque necrosis.
The Journal of clinical investigation
Doran AC,Ozcan L,Cai B,Zheng Z,Fredman G,Rymond CC,Dorweiler B,Sluimer JC,Hsieh J,Kuriakose G,Tall AR,Tabas I
Wed Nov 01 00:00:00 EDT 2017
CAMKIIγ suppresses an efferocytosis pathway in macrophages and promotes atherosclerotic plaque necrosis.
The Journal of clinical investigation
Doran AC,Ozcan L,Cai B,Zheng Z,Fredman G,Rymond CC,Dorweiler B,Sluimer JC,Hsieh J,Kuriakose G,Tall AR,Tabas I
Wed Nov 01 00:00:00 EDT 2017
Alternative Progenitor Cells Compensate to Rebuild the Coronary Vasculature in Elabela- and Apj-Deficient Hearts.
Developmental cell
Sharma B,Ho L,Ford GH,Chen HI,Goldstone AB,Woo YJ,Quertermous T,Reversade B,Red-Horse K
Mon Sep 25 00:00:00 EDT 2017
Alternative Progenitor Cells Compensate to Rebuild the Coronary Vasculature in Elabela- and Apj-Deficient Hearts.
Developmental cell
Sharma B,Ho L,Ford GH,Chen HI,Goldstone AB,Woo YJ,Quertermous T,Reversade B,Red-Horse K
Mon Sep 25 00:00:00 EDT 2017
DACH1 stimulates shear stress-guided endothelial cell migration and coronary artery growth through the CXCL12-CXCR4 signaling axis.
Genes & development
Chang AH,Raftrey BC,D'Amato G,Surya VN,Poduri A,Chen HI,Goldstone AB,Woo J,Fuller GG,Dunn AR,Red-Horse K
Sat Jul 01 00:00:00 EDT 2017
DACH1 stimulates shear stress-guided endothelial cell migration and coronary artery growth through the CXCL12-CXCR4 signaling axis.
Genes & development
Chang AH,Raftrey BC,D'Amato G,Surya VN,Poduri A,Chen HI,Goldstone AB,Woo J,Fuller GG,Dunn AR,Red-Horse K
Sat Jul 01 00:00:00 EDT 2017
Human iPSC-Derived Neural Progenitors Are an Effective Drug Discovery Model for Neurological mtDNA Disorders.
Cell stem cell
Lorenz C,Lesimple P,Bukowiecki R,Zink A,Inak G,Mlody B,Singh M,Semtner M,Mah N,Auré K,Leong M,Zabiegalov O,Lyras EM,Pfiffer V,Fauler B,Eichhorst J,Wiesner B,Huebner N,Priller J,Mielke T,Meierhofer D,Izsvák Z,Meier JC,Bouillaud F,Adjaye J,Schuelke M,Wanker EE,Lombès A,Prigione A
Thu May 04 00:00:00 EDT 2017
Human iPSC-Derived Neural Progenitors Are an Effective Drug Discovery Model for Neurological mtDNA Disorders.
Cell stem cell
Lorenz C,Lesimple P,Bukowiecki R,Zink A,Inak G,Mlody B,Singh M,Semtner M,Mah N,Auré K,Leong M,Zabiegalov O,Lyras EM,Pfiffer V,Fauler B,Eichhorst J,Wiesner B,Huebner N,Priller J,Mielke T,Meierhofer D,Izsvák Z,Meier JC,Bouillaud F,Adjaye J,Schuelke M,Wanker EE,Lombès A,Prigione A
Thu May 04 00:00:00 EDT 2017
DACH1 suppresses breast cancer as a negative regulator of CD44.
Scientific reports
Xu H,Yu S,Yuan X,Xiong J,Kuang D,Pestell RG,Wu K
Wed Jun 28 00:00:00 EDT 2017
DACH1 suppresses breast cancer as a negative regulator of CD44.
Scientific reports
Xu H,Yu S,Yuan X,Xiong J,Kuang D,Pestell RG,Wu K
Wed Jun 28 00:00:00 EDT 2017
YB-1 regulates tiRNA-induced Stress Granule formation but not translational repression.
Nucleic acids research
Lyons SM,Achorn C,Kedersha NL,Anderson PJ,Ivanov P
Fri Aug 19 00:00:00 EDT 2016
YB-1 regulates tiRNA-induced Stress Granule formation but not translational repression.
Nucleic acids research
Lyons SM,Achorn C,Kedersha NL,Anderson PJ,Ivanov P
Fri Aug 19 00:00:00 EDT 2016
Partial Dedifferentiation of Murine Radial Glia-Type Neural Stem Cells by Brn2 and c-Myc Yields Early Neuroepithelial Progenitors.
Journal of molecular biology
Bung R,Wörsdörfer P,Thier MC,Lemke K,Gebhardt M,Edenhofer F
Sun Apr 10 00:00:00 EDT 2016
Partial Dedifferentiation of Murine Radial Glia-Type Neural Stem Cells by Brn2 and c-Myc Yields Early Neuroepithelial Progenitors.
Journal of molecular biology
Bung R,Wörsdörfer P,Thier MC,Lemke K,Gebhardt M,Edenhofer F
Sun Apr 10 00:00:00 EDT 2016
Inhibition of DACH1 activity by short hairpin RNA represses cell proliferation and tumor invasion in pancreatic cancer.
Oncology reports
Bu XN,Qiu C,Wang C,Jiang Z
Mon Aug 01 00:00:00 EDT 2016
Robust production of human neural cells by establishing neuroepithelial-like stem cells from peripheral blood mononuclear cell-derived feeder-free iPSCs under xeno-free conditions.
Neuroscience research
Isoda M,Kohyama J,Iwanami A,Sanosaka T,Sugai K,Yamaguchi R,Matsumoto T,Nakamura M,Okano H
Thu Sep 01 00:00:00 EDT 2016
Robust production of human neural cells by establishing neuroepithelial-like stem cells from peripheral blood mononuclear cell-derived feeder-free iPSCs under xeno-free conditions.
Neuroscience research
Isoda M,Kohyama J,Iwanami A,Sanosaka T,Sugai K,Yamaguchi R,Matsumoto T,Nakamura M,Okano H
Thu Sep 01 00:00:00 EDT 2016
Decreased DACH1 expression in glomerulopathy is associated with disease progression and severity.
Oncotarget
Liu QQ,Zhou YQ,Liu HQ,Qiu WH,Liu H,Hu TY,Xu Q,Lv YM,Wu KM
Tue Dec 27 00:00:00 EST 2016
Decreased DACH1 expression in glomerulopathy is associated with disease progression and severity.
Oncotarget
Liu QQ,Zhou YQ,Liu HQ,Qiu WH,Liu H,Hu TY,Xu Q,Lv YM,Wu KM
Tue Dec 27 00:00:00 EST 2016
Decreased DACH1 expression in glomerulopathy is associated with disease progression and severity.
Oncotarget
Liu QQ,Zhou YQ,Liu HQ,Qiu WH,Liu H,Hu TY,Xu Q,Lv YM,Wu KM
Tue Dec 27 00:00:00 EST 2016
Specific Inhibition of β-Secretase Processing of the Alzheimer Disease Amyloid Precursor Protein.
Cell reports
Ben Halima S,Mishra S,Raja KMP,Willem M,Baici A,Simons K,Brüstle O,Koch P,Haass C,Caflisch A,Rajendran L
Tue Mar 08 00:00:00 EST 2016
Specific Inhibition of β-Secretase Processing of the Alzheimer Disease Amyloid Precursor Protein.
Cell reports
Ben Halima S,Mishra S,Raja KMP,Willem M,Baici A,Simons K,Brüstle O,Koch P,Haass C,Caflisch A,Rajendran L
Tue Mar 08 00:00:00 EST 2016
The endogenous cell-fate factor dachshund restrains prostate epithelial cell migration via repression of cytokine secretion via a cxcl signaling module.
Cancer research
Chen K,Wu K,Jiao X,Wang L,Ju X,Wang M,Di Sante G,Xu S,Wang Q,Li K,Sun X,Xu C,Li Z,Casimiro MC,Ertel A,Addya S,McCue PA,Lisanti MP,Wang C,Davis RJ,Mardon G,Pestell RG
Fri May 15 00:00:00 EDT 2015
The endogenous cell-fate factor dachshund restrains prostate epithelial cell migration via repression of cytokine secretion via a cxcl signaling module.
Cancer research
Chen K,Wu K,Jiao X,Wang L,Ju X,Wang M,Di Sante G,Xu S,Wang Q,Li K,Sun X,Xu C,Li Z,Casimiro MC,Ertel A,Addya S,McCue PA,Lisanti MP,Wang C,Davis RJ,Mardon G,Pestell RG
Fri May 15 00:00:00 EDT 2015
The endogenous cell-fate factor dachshund restrains prostate epithelial cell migration via repression of cytokine secretion via a cxcl signaling module.
Cancer research
Chen K,Wu K,Jiao X,Wang L,Ju X,Wang M,Di Sante G,Xu S,Wang Q,Li K,Sun X,Xu C,Li Z,Casimiro MC,Ertel A,Addya S,McCue PA,Lisanti MP,Wang C,Davis RJ,Mardon G,Pestell RG
Fri May 15 00:00:00 EDT 2015
The endogenous cell-fate factor dachshund restrains prostate epithelial cell migration via repression of cytokine secretion via a cxcl signaling module.
Cancer research
Chen K,Wu K,Jiao X,Wang L,Ju X,Wang M,Di Sante G,Xu S,Wang Q,Li K,Sun X,Xu C,Li Z,Casimiro MC,Ertel A,Addya S,McCue PA,Lisanti MP,Wang C,Davis RJ,Mardon G,Pestell RG
Fri May 15 00:00:00 EDT 2015
Estrogen receptor β exon 3-deleted mouse: The importance of non-ERE pathways in ERβ signaling.
Proceedings of the National Academy of Sciences of the United States of America
Maneix L,Antonson P,Humire P,Rochel-Maia S,Castañeda J,Omoto Y,Kim HJ,Warner M,Gustafsson JÅ
Tue Apr 21 00:00:00 EDT 2015
Estrogen receptor β exon 3-deleted mouse: The importance of non-ERE pathways in ERβ signaling.
Proceedings of the National Academy of Sciences of the United States of America
Maneix L,Antonson P,Humire P,Rochel-Maia S,Castañeda J,Omoto Y,Kim HJ,Warner M,Gustafsson JÅ
Tue Apr 21 00:00:00 EDT 2015
Estrogen receptor β exon 3-deleted mouse: The importance of non-ERE pathways in ERβ signaling.
Proceedings of the National Academy of Sciences of the United States of America
Maneix L,Antonson P,Humire P,Rochel-Maia S,Castañeda J,Omoto Y,Kim HJ,Warner M,Gustafsson JÅ
Tue Apr 21 00:00:00 EDT 2015
Estrogen receptor β exon 3-deleted mouse: The importance of non-ERE pathways in ERβ signaling.
Proceedings of the National Academy of Sciences of the United States of America
Maneix L,Antonson P,Humire P,Rochel-Maia S,Castañeda J,Omoto Y,Kim HJ,Warner M,Gustafsson JÅ
Tue Apr 21 00:00:00 EDT 2015
Androgen therapy reverses injuries caused by ethanol consumption in the prostate: testosterone as a possible target to ethanol-related disorders.
Life sciences
Mendes LO,Scarano WR,Rochel-Maia SS,Fioruci-Fontaneli BA,Chuffa LG,Anselmo-Franci JA,Martinez FE
Thu Jan 01 00:00:00 EST 2015
Androgen therapy reverses injuries caused by ethanol consumption in the prostate: testosterone as a possible target to ethanol-related disorders.
Life sciences
Mendes LO,Scarano WR,Rochel-Maia SS,Fioruci-Fontaneli BA,Chuffa LG,Anselmo-Franci JA,Martinez FE
Thu Jan 01 00:00:00 EST 2015
Androgen therapy reverses injuries caused by ethanol consumption in the prostate: testosterone as a possible target to ethanol-related disorders.
Life sciences
Mendes LO,Scarano WR,Rochel-Maia SS,Fioruci-Fontaneli BA,Chuffa LG,Anselmo-Franci JA,Martinez FE
Thu Jan 01 00:00:00 EST 2015
Androgen therapy reverses injuries caused by ethanol consumption in the prostate: testosterone as a possible target to ethanol-related disorders.
Life sciences
Mendes LO,Scarano WR,Rochel-Maia SS,Fioruci-Fontaneli BA,Chuffa LG,Anselmo-Franci JA,Martinez FE
Thu Jan 01 00:00:00 EST 2015
DACH1 is a novel predictive and prognostic biomarker in hepatocellular carcinoma as a negative regulator of Wnt/β-catenin signaling.
Oncotarget
Liu Y,Zhou R,Yuan X,Han N,Zhou S,Xu H,Guo M,Yu S,Zhang C,Yin T,Wu K
Mon Apr 20 00:00:00 EDT 2015
DACH1 is a novel predictive and prognostic biomarker in hepatocellular carcinoma as a negative regulator of Wnt/β-catenin signaling.
Oncotarget
Liu Y,Zhou R,Yuan X,Han N,Zhou S,Xu H,Guo M,Yu S,Zhang C,Yin T,Wu K
Mon Apr 20 00:00:00 EDT 2015
DACH1 is a novel predictive and prognostic biomarker in hepatocellular carcinoma as a negative regulator of Wnt/β-catenin signaling.
Oncotarget
Liu Y,Zhou R,Yuan X,Han N,Zhou S,Xu H,Guo M,Yu S,Zhang C,Yin T,Wu K
Mon Apr 20 00:00:00 EDT 2015
DACH1 is a novel predictive and prognostic biomarker in hepatocellular carcinoma as a negative regulator of Wnt/β-catenin signaling.
Oncotarget
Liu Y,Zhou R,Yuan X,Han N,Zhou S,Xu H,Guo M,Yu S,Zhang C,Yin T,Wu K
Mon Apr 20 00:00:00 EDT 2015
DACH1 inhibits lung adenocarcinoma invasion and tumor growth by repressing CXCL5 signaling.
Oncotarget
Han N,Yuan X,Wu H,Xu H,Chu Q,Guo M,Yu S,Chen Y,Wu K
Fri Mar 20 00:00:00 EDT 2015
DACH1 inhibits lung adenocarcinoma invasion and tumor growth by repressing CXCL5 signaling.
Oncotarget
Han N,Yuan X,Wu H,Xu H,Chu Q,Guo M,Yu S,Chen Y,Wu K
Fri Mar 20 00:00:00 EDT 2015
DACH1 inhibits lung adenocarcinoma invasion and tumor growth by repressing CXCL5 signaling.
Oncotarget
Han N,Yuan X,Wu H,Xu H,Chu Q,Guo M,Yu S,Chen Y,Wu K
Fri Mar 20 00:00:00 EDT 2015
Arylsulfatase A Overexpressing Human iPSC-derived Neural Cells Reduce CNS Sulfatide Storage in a Mouse Model of Metachromatic Leukodystrophy.
Molecular therapy : the journal of the American Society of Gene Therapy
Doerr J,Böckenhoff A,Ewald B,Ladewig J,Eckhardt M,Gieselmann V,Matzner U,Brüstle O,Koch P
Tue Sep 01 00:00:00 EDT 2015
Auto-attraction of neural precursors and their neuronal progeny impairs neuronal migration.
Nature neuroscience
Ladewig J,Koch P,Brüstle O
Wed Jan 01 00:00:00 EST 2014
Auto-attraction of neural precursors and their neuronal progeny impairs neuronal migration.
Nature neuroscience
Ladewig J,Koch P,Brüstle O
Wed Jan 01 00:00:00 EST 2014
The sinus venosus contributes to coronary vasculature through VEGFC-stimulated angiogenesis.
Development (Cambridge, England)
Chen HI,Sharma B,Akerberg BN,Numi HJ,Kivelä R,Saharinen P,Aghajanian H,McKay AS,Bogard PE,Chang AH,Jacobs AH,Epstein JA,Stankunas K,Alitalo K,Red-Horse K
Mon Dec 01 00:00:00 EST 2014
The sinus venosus contributes to coronary vasculature through VEGFC-stimulated angiogenesis.
Development (Cambridge, England)
Chen HI,Sharma B,Akerberg BN,Numi HJ,Kivelä R,Saharinen P,Aghajanian H,McKay AS,Bogard PE,Chang AH,Jacobs AH,Epstein JA,Stankunas K,Alitalo K,Red-Horse K
Mon Dec 01 00:00:00 EST 2014
Epigenetic silencing of DACH1 induces the invasion and metastasis of gastric cancer by activating TGF-β signalling.
Journal of cellular and molecular medicine
Yan W,Wu K,Herman JG,Brock MV,Zhou Y,Lu Y,Zhang Z,Yang Y,Guo M
Mon Dec 01 00:00:00 EST 2014
Epigenetic silencing of DACH1 induces the invasion and metastasis of gastric cancer by activating TGF-β signalling.
Journal of cellular and molecular medicine
Yan W,Wu K,Herman JG,Brock MV,Zhou Y,Lu Y,Zhang Z,Yang Y,Guo M
Mon Dec 01 00:00:00 EST 2014
Epigenetic silencing of DACH1 induces the invasion and metastasis of gastric cancer by activating TGF-β signalling.
Journal of cellular and molecular medicine
Yan W,Wu K,Herman JG,Brock MV,Zhou Y,Lu Y,Zhang Z,Yang Y,Guo M
Mon Dec 01 00:00:00 EST 2014
Epigenetic silencing of DACH1 induces the invasion and metastasis of gastric cancer by activating TGF-β signalling.
Journal of cellular and molecular medicine
Yan W,Wu K,Herman JG,Brock MV,Zhou Y,Lu Y,Zhang Z,Yang Y,Guo M
Mon Dec 01 00:00:00 EST 2014
Silencing DACH1 promotes esophageal cancer growth by inhibiting TGF-β signaling.
PloS one
Wu L,Herman JG,Brock MV,Wu K,Mao G,Yan W,Nie Y,Liang H,Zhan Q,Li W,Guo M
Fri Jun 05 00:00:00 EDT 2015
Silencing DACH1 promotes esophageal cancer growth by inhibiting TGF-β signaling.
PloS one
Wu L,Herman JG,Brock MV,Wu K,Mao G,Yan W,Nie Y,Liang H,Zhan Q,Li W,Guo M
Fri Jun 05 00:00:00 EDT 2015
Silencing DACH1 promotes esophageal cancer growth by inhibiting TGF-β signaling.
PloS one
Wu L,Herman JG,Brock MV,Wu K,Mao G,Yan W,Nie Y,Liang H,Zhan Q,Li W,Guo M
Fri Jun 05 00:00:00 EDT 2015
Silencing DACH1 promotes esophageal cancer growth by inhibiting TGF-β signaling.
PloS one
Wu L,Herman JG,Brock MV,Wu K,Mao G,Yan W,Nie Y,Liang H,Zhan Q,Li W,Guo M
Fri Jun 05 00:00:00 EDT 2015
DACH1 inhibits cyclin D1 expression, cellular proliferation and tumor growth of renal cancer cells.
Journal of hematology & oncology
Chu Q,Han N,Yuan X,Nie X,Wu H,Chen Y,Guo M,Yu S,Wu K
Fri Oct 17 00:00:00 EDT 2014
DACH1 inhibits cyclin D1 expression, cellular proliferation and tumor growth of renal cancer cells.
Journal of hematology & oncology
Chu Q,Han N,Yuan X,Nie X,Wu H,Chen Y,Guo M,Yu S,Wu K
Fri Oct 17 00:00:00 EDT 2014
DACH1 inhibits cyclin D1 expression, cellular proliferation and tumor growth of renal cancer cells.
Journal of hematology & oncology
Chu Q,Han N,Yuan X,Nie X,Wu H,Chen Y,Guo M,Yu S,Wu K
Fri Oct 17 00:00:00 EDT 2014
DACH1 inhibits cyclin D1 expression, cellular proliferation and tumor growth of renal cancer cells.
Journal of hematology & oncology
Chu Q,Han N,Yuan X,Nie X,Wu H,Chen Y,Guo M,Yu S,Wu K
Fri Oct 17 00:00:00 EDT 2014
Dachshund binds p53 to block the growth of lung adenocarcinoma cells.
Cancer research
Chen K,Wu K,Cai S,Zhang W,Zhou J,Wang J,Ertel A,Li Z,Rui H,Quong A,Lisanti MP,Tozeren A,Tanes C,Addya S,Gormley M,Wang C,McMahon SB,Pestell RG
Sat Jun 01 00:00:00 EDT 2013
Dachshund binds p53 to block the growth of lung adenocarcinoma cells.
Cancer research
Chen K,Wu K,Cai S,Zhang W,Zhou J,Wang J,Ertel A,Li Z,Rui H,Quong A,Lisanti MP,Tozeren A,Tanes C,Addya S,Gormley M,Wang C,McMahon SB,Pestell RG
Sat Jun 01 00:00:00 EDT 2013
Dachshund binds p53 to block the growth of lung adenocarcinoma cells.
Cancer research
Chen K,Wu K,Cai S,Zhang W,Zhou J,Wang J,Ertel A,Li Z,Rui H,Quong A,Lisanti MP,Tozeren A,Tanes C,Addya S,Gormley M,Wang C,McMahon SB,Pestell RG
Sat Jun 01 00:00:00 EDT 2013
Dachshund binds p53 to block the growth of lung adenocarcinoma cells.
Cancer research
Chen K,Wu K,Cai S,Zhang W,Zhou J,Wang J,Ertel A,Li Z,Rui H,Quong A,Lisanti MP,Tozeren A,Tanes C,Addya S,Gormley M,Wang C,McMahon SB,Pestell RG
Sat Jun 01 00:00:00 EDT 2013
Dachshund binds p53 to block the growth of lung adenocarcinoma cells.
Cancer research
Chen K,Wu K,Cai S,Zhang W,Zhou J,Wang J,Ertel A,Li Z,Rui H,Quong A,Lisanti MP,Tozeren A,Tanes C,Addya S,Gormley M,Wang C,McMahon SB,Pestell RG
Sat Jun 01 00:00:00 EDT 2013
APP processing in human pluripotent stem cell-derived neurons is resistant to NSAID-based γ-secretase modulation.
Stem cell reports
Mertens J,Stüber K,Wunderlich P,Ladewig J,Kesavan JC,Vandenberghe R,Vandenbulcke M,van Damme P,Walter J,Brüstle O,Koch P
Mon May 11 00:00:00 EDT 2015
APP processing in human pluripotent stem cell-derived neurons is resistant to NSAID-based γ-secretase modulation.
Stem cell reports
Mertens J,Stüber K,Wunderlich P,Ladewig J,Kesavan JC,Vandenberghe R,Vandenbulcke M,van Damme P,Walter J,Brüstle O,Koch P
Mon May 11 00:00:00 EDT 2015
Stem cells expanded from the human embryonic hindbrain stably retain regional specification and high neurogenic potency.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Tailor J,Kittappa R,Leto K,Gates M,Borel M,Paulsen O,Spitzer S,Karadottir RT,Rossi F,Falk A,Smith A
Wed Jul 24 00:00:00 EDT 2013
Stem cells expanded from the human embryonic hindbrain stably retain regional specification and high neurogenic potency.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Tailor J,Kittappa R,Leto K,Gates M,Borel M,Paulsen O,Spitzer S,Karadottir RT,Rossi F,Falk A,Smith A
Wed Jul 24 00:00:00 EDT 2013
Embryonic stem cell-based modeling of tau pathology in human neurons.
The American journal of pathology
Mertens J,Stüber K,Poppe D,Doerr J,Ladewig J,Brüstle O,Koch P
Wed May 01 00:00:00 EDT 2013
Embryonic stem cell-based modeling of tau pathology in human neurons.
The American journal of pathology
Mertens J,Stüber K,Poppe D,Doerr J,Ladewig J,Brüstle O,Koch P
Wed May 01 00:00:00 EDT 2013
Epigenetic regulation of DACH1, a novel Wnt signaling component in colorectal cancer.
Epigenetics
Yan W,Wu K,Herman JG,Brock MV,Fuks F,Yang L,Zhu H,Li Y,Yang Y,Guo M
Sun Dec 01 00:00:00 EST 2013
Epigenetic regulation of DACH1, a novel Wnt signaling component in colorectal cancer.
Epigenetics
Yan W,Wu K,Herman JG,Brock MV,Fuks F,Yang L,Zhu H,Li Y,Yang Y,Guo M
Sun Dec 01 00:00:00 EST 2013
Specification of functional cranial placode derivatives from human pluripotent stem cells.
Cell reports
Dincer Z,Piao J,Niu L,Ganat Y,Kriks S,Zimmer B,Shi SH,Tabar V,Studer L
Thu Dec 12 00:00:00 EST 2013
Specification of functional cranial placode derivatives from human pluripotent stem cells.
Cell reports
Dincer Z,Piao J,Niu L,Ganat Y,Kriks S,Zimmer B,Shi SH,Tabar V,Studer L
Thu Dec 12 00:00:00 EST 2013
MicroRNA-195 targets ADP-ribosylation factor-like protein 2 to induce apoptosis in human embryonic stem cell-derived neural progenitor cells.
Cell death & disease
Zhou Y,Jiang H,Gu J,Tang Y,Shen N,Jin Y
Thu Jun 27 00:00:00 EDT 2013
MicroRNA-195 targets ADP-ribosylation factor-like protein 2 to induce apoptosis in human embryonic stem cell-derived neural progenitor cells.
Cell death & disease
Zhou Y,Jiang H,Gu J,Tang Y,Shen N,Jin Y
Thu Jun 27 00:00:00 EDT 2013
Excitation-induced ataxin-3 aggregation in neurons from patients with Machado-Joseph disease.
Nature
Koch P,Breuer P,Peitz M,Jungverdorben J,Kesavan J,Poppe D,Doerr J,Ladewig J,Mertens J,Tüting T,Hoffmann P,Klockgether T,Evert BO,Wüllner U,Brüstle O
Wed Nov 23 00:00:00 EST 2011
Excitation-induced ataxin-3 aggregation in neurons from patients with Machado-Joseph disease.
Nature
Koch P,Breuer P,Peitz M,Jungverdorben J,Kesavan J,Poppe D,Doerr J,Ladewig J,Mertens J,Tüting T,Hoffmann P,Klockgether T,Evert BO,Wüllner U,Brüstle O
Wed Nov 23 00:00:00 EST 2011
Dachshund homologues play a conserved role in islet cell development.
Developmental biology
Kalousova A,Mavropoulos A,Adams BA,Nekrep N,Li Z,Krauss S,Stainier DY,German MS
Wed Dec 15 00:00:00 EST 2010
Dachshund homologues play a conserved role in islet cell development.
Developmental biology
Kalousova A,Mavropoulos A,Adams BA,Nekrep N,Li Z,Krauss S,Stainier DY,German MS
Wed Dec 15 00:00:00 EST 2010
Dachshund homologues play a conserved role in islet cell development.
Developmental biology
Kalousova A,Mavropoulos A,Adams BA,Nekrep N,Li Z,Krauss S,Stainier DY,German MS
Wed Dec 15 00:00:00 EST 2010
Dachshund homologues play a conserved role in islet cell development.
Developmental biology
Kalousova A,Mavropoulos A,Adams BA,Nekrep N,Li Z,Krauss S,Stainier DY,German MS
Wed Dec 15 00:00:00 EST 2010
A rosette-type, self-renewing human ES cell-derived neural stem cell with potential for in vitro instruction and synaptic integration.
Proceedings of the National Academy of Sciences of the United States of America
Koch P,Opitz T,Steinbeck JA,Ladewig J,Brüstle O
Tue Mar 03 00:00:00 EST 2009
A rosette-type, self-renewing human ES cell-derived neural stem cell with potential for in vitro instruction and synaptic integration.
Proceedings of the National Academy of Sciences of the United States of America
Koch P,Opitz T,Steinbeck JA,Ladewig J,Brüstle O
Tue Mar 03 00:00:00 EST 2009
Human ES cell-derived neural rosettes reveal a functionally distinct early neural stem cell stage.
Genes & development
Elkabetz Y,Panagiotakos G,Al Shamy G,Socci ND,Tabar V,Studer L
Tue Jan 15 00:00:00 EST 2008
Human ES cell-derived neural rosettes reveal a functionally distinct early neural stem cell stage.
Genes & development
Elkabetz Y,Panagiotakos G,Al Shamy G,Socci ND,Tabar V,Studer L
Tue Jan 15 00:00:00 EST 2008