Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor, beta subunit 1; GA-binding protein subunit beta-1; GABP subunit beta-1; GABP subunit beta-2; GABPB-2; Nuclear respiratory factor 2; Transcription factor E4TF1-47; Transcription factor E4TF1-53
Gene Aliases: BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4TF1B; GABPB; GABPB-1; GABPB1; GABPB2; NRF2B1; NRF2B2
UniProt ID: (Human) Q06547
Entrez Gene ID: (Human) 2553
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support