Product References
Tip60/KAT5 Histone Acetyltransferase Is Required for Maintenance and Neurogenesis of Embryonic Neural Stem Cells.
International journal of molecular sciences
Tominaga K,Sakashita E,Kasashima K,Kuroiwa K,Nagao Y,Iwamori N,Endo H
Fri Jan 20 00:00:00 EST 2023
Role of miR-219a-5p in regulating NMDAR in nonylphenol-induced synaptic plasticity damage.
Ecotoxicology and environmental safety
Fu N,Yu J,Zhu L,Yang L,Ma L,He J,Yu H,Liu J,Tian Y,Xu J
Wed Mar 01 00:00:00 EST 2023
The collateral activity of RfxCas13d can induce lethality in a RfxCas13d knock-in mouse model.
Genome biology
Li Y,Xu J,Guo X,Li Z,Cao L,Liu S,Guo Y,Wang G,Luo Y,Zhang Z,Wei X,Zhao Y,Liu T,Wang X,Xia H,Kuang M,Guo Q,Li J,Chen L,Wang Y,Li Q,Wang F,Liu Q,You F
Wed Feb 01 00:00:00 EST 2023
DLP printed hDPSC-loaded GelMA microsphere regenerates dental pulp and repairs spinal cord.
Biomaterials
Qian Y,Gong J,Lu K,Hong Y,Zhu Z,Zhang J,Zou Y,Zhou F,Zhang C,Zhou S,Gu T,Sun M,Wang S,He J,Li Y,Lin J,Yuan Y,Ouyang H,Yu M,Wang H
Tue Aug 01 00:00:00 EDT 2023
A whole transcriptome profiling analysis for antidepressant mechanism of Xiaoyaosan mediated synapse loss via BDNF/trkB/PI3K signal axis in CUMS rats.
BMC complementary medicine and therapies
Meng P,Zhang X,Liu TT,Liu J,Luo Y,Xie MX,Yang H,Fang R,Guo DW,Zhong ZY,Wang YH,Ge JW
Thu Jun 15 00:00:00 EDT 2023
Microglia PKM2 Mediates Neuroinflammation and Neuron Loss in Mice Epilepsy through the Astrocyte C3-Neuron C3R Signaling Pathway.
Brain sciences
Li X,Zhou R,Peng H,Peng J,Li Q,Mei M
Fri Feb 03 00:00:00 EST 2023
A novel small-molecular CCR5 antagonist promotes neural repair after stroke.
Acta pharmacologica Sinica
Wu QL,Cui LY,Ma WY,Wang SS,Zhang Z,Feng ZP,Sun HS,Chu SF,He WB,Chen NH
Sun Oct 01 00:00:00 EDT 2023
20S-Protopanaxatriol improves cognitive function of Alzheimer's disease by promoting endogenous neurogenesis.
Food & function
Zhang Q,Yang X,Liu J,Song J,Zhang S,Chen L,Zhang M
Thu May 11 00:00:00 EDT 2023
Transient neurogenesis in ischemic cortex from Sox2+ astrocytes.
Neural regeneration research
Yang JL,Fan H,Fu FF,Guo BL,Huang Y,Sun L,Wang WT,Xing JL,Hu XT,Ding YQ,Zhang K,Hu YZ,Wang YZ
Sat Jul 01 00:00:00 EDT 2023
Primary cilia TRP channel regulates hippocampal excitability.
Proceedings of the National Academy of Sciences of the United States of America
Vien TN,Ta MC,Kimura LF,Onay T,DeCaen PG
Tue May 30 00:00:00 EDT 2023
Exposure to bisphenol A induces neurotoxicity associated with synaptic and cytoskeletal dysfunction in neuro-2a cells.
Toxicology and industrial health
Wang S,Ning H,Wang X,Chen L,Hua L,Ren F,Hu D,Li R,Ma Z,Ge Y,Yin Z
Thu Jun 01 00:00:00 EDT 2023
Deregulation of ER-mitochondria contact formation and mitochondrial calcium homeostasis mediated by VDAC in fragile X syndrome.
Developmental cell
Geng J,Khaket TP,Pan J,Li W,Zhang Y,Ping Y,Cobos Sillero MI,Lu B
Mon Apr 10 00:00:00 EDT 2023
Deregulation of ER-mitochondria contact formation and mitochondrial calcium homeostasis mediated by VDAC in fragile X syndrome.
Developmental cell
Geng J,Khaket TP,Pan J,Li W,Zhang Y,Ping Y,Cobos Sillero MI,Lu B
Mon Apr 10 00:00:00 EDT 2023
Taxifolin attenuates neuroinflammation and microglial pyroptosis via the PI3K/Akt signaling pathway after spinal cord injury.
International immunopharmacology
Hu Z,Xuan L,Wu T,Jiang N,Liu X,Chang J,Wang T,Han N,Tian X
Sun Jan 01 00:00:00 EST 2023
Taxifolin attenuates neuroinflammation and microglial pyroptosis via the PI3K/Akt signaling pathway after spinal cord injury.
International immunopharmacology
Hu Z,Xuan L,Wu T,Jiang N,Liu X,Chang J,Wang T,Han N,Tian X
Sun Jan 01 00:00:00 EST 2023
Taxifolin attenuates neuroinflammation and microglial pyroptosis via the PI3K/Akt signaling pathway after spinal cord injury.
International immunopharmacology
Hu Z,Xuan L,Wu T,Jiang N,Liu X,Chang J,Wang T,Han N,Tian X
Sun Jan 01 00:00:00 EST 2023
Taxifolin attenuates neuroinflammation and microglial pyroptosis via the PI3K/Akt signaling pathway after spinal cord injury.
International immunopharmacology
Hu Z,Xuan L,Wu T,Jiang N,Liu X,Chang J,Wang T,Han N,Tian X
Sun Jan 01 00:00:00 EST 2023
Effects of early-life tosufloxacin tosilate hydrate administration on growth rate, neurobehavior, and gut microbiota at adulthood in male mice.
The Journal of toxicological sciences
Hasegawa A,Sasaki T,Islam J,Tominaga T,Nochi T,Hara K,Tanemura K
Fri Mar 03 00:00:00 EST 2023
Peroxiredoxin-6 Released by Astrocytes Contributes to Neuroapoptosis During Ischemia.
Neuroscience
Hou JY,Zhou XL,Wang XY,Liang J,Xue Q
Tue Feb 21 00:00:00 EST 2023
Effects of magnetically targeted iron oxide@polydopamine-labeled human umbilical cord mesenchymal stem cells in cerebral infarction in mice.
Aging
Yan J,Liu T,Li Y,Zhang J,Shi B,Zhang F,Hou X,Zhang X,Cui W,Li J,Yao H,Li X,Gao Y,Jiang J
Tue Feb 28 00:00:00 EST 2023
Kynurenic acid blunts A1 astrocyte activation against neurodegeneration in HIV-associated neurocognitive disorders.
Journal of neuroinflammation
Lun J,Li Y,Gao X,Gong Z,Chen X,Zou J,Zhou C,Huang Y,Zhou B,Huang P,Cao H
Thu Mar 30 00:00:00 EDT 2023
Kynurenic acid blunts A1 astrocyte activation against neurodegeneration in HIV-associated neurocognitive disorders.
Journal of neuroinflammation
Lun J,Li Y,Gao X,Gong Z,Chen X,Zou J,Zhou C,Huang Y,Zhou B,Huang P,Cao H
Thu Mar 30 00:00:00 EDT 2023
MiR-29c Inhibits TNF-α-Induced ROS Production and Apoptosis in Mouse Hippocampal HT22 Cell Line.
Neurochemical research
Li B,Lu Y,Wang R,Xu T,Lei X,Jin H,Gao X,Xie Y,Liu X,Zeng J
Wed Feb 01 00:00:00 EST 2023
Golgi damage caused by dysfunction of PiT-2 in primary familial brain calcification.
Biochemical and biophysical research communications
Sun H,Wang Z,Zhang Q,Chen N,Tang M,Yang Z,Xu Y,Kang J,Wang Y
Sun Jan 29 00:00:00 EST 2023
Senile plaques in Alzheimer's disease arise from Aβ- and Cathepsin D-enriched mixtures leaking out during intravascular haemolysis and microaneurysm rupture.
FEBS letters
Fu H,Li J,Du P,Jin W,Gao G,Cui D
Sat Apr 01 00:00:00 EDT 2023
Transmission of NLRP3-IL-1β Signals in Cerebral Ischemia and Reperfusion Injury: from Microglia to Adjacent Neuron and Endothelial Cells via IL-1β/IL-1R1/TRAF6.
Molecular neurobiology
Pan J,Peng J,Li X,Wang H,Rong X,Peng Y
Mon May 01 00:00:00 EDT 2023
The total alkaloids of Sophora alopecuroides L. improve depression-like behavior in mice via BDNF-mediated AKT/mTOR signaling pathway.
Journal of ethnopharmacology
Li J,Zhang M,Pei Y,Yang Q,Zheng L,Wang G,Sun Y,Yang W,Liu L
Wed Nov 15 00:00:00 EST 2023
Overexpression of vascular endothelial growth factor enhances the neuroprotective effects of bone marrow mesenchymal stem cell transplantation in ischemic stroke.
Neural regeneration research
Liu C,Yang ZX,Zhou SQ,Ding D,Hu YT,Yang HN,Han D,Hu SQ,Zong XM
Thu Jun 01 00:00:00 EDT 2023
Overexpression of vascular endothelial growth factor enhances the neuroprotective effects of bone marrow mesenchymal stem cell transplantation in ischemic stroke.
Neural regeneration research
Liu C,Yang ZX,Zhou SQ,Ding D,Hu YT,Yang HN,Han D,Hu SQ,Zong XM
Thu Jun 01 00:00:00 EDT 2023
miR-101a-3p Impairs Synaptic Plasticity and Contributes to Synucleinopathy.
Journal of Parkinson's disease
Xylaki M,Paiva I,Al-Azzani M,Gerhardt E,Jain G,Islam MR,Vasili E,Wassouf Z,Schulze-Hentrich JM,Fischer A,Outeiro TF
Tue Mar 21 00:00:00 EDT 2023
Cofilin promotes tau pathology in Alzheimer's disease.
Cell reports
Yan M,Tang L,Dai L,Lei C,Xiong M,Zhang X,He M,Tian Y,Xiong J,Ke W,Zhang Z,Zhang C,Deng X,Zhang Z
Tue Feb 28 00:00:00 EST 2023
Mitochondrial transplantation ameliorates hippocampal damage following status epilepticus.
Animal models and experimental medicine
Jia X,Wang Q,Ji J,Lu W,Liu Z,Tian H,Guo L,Wang Y
Wed Feb 01 00:00:00 EST 2023
Engineered extracellular vesicles for delivery of siRNA promoting targeted repair of traumatic spinal cord injury.
Bioactive materials
Rong Y,Wang Z,Tang P,Wang J,Ji C,Chang J,Zhu Y,Ye W,Bai J,Liu W,Yin G,Yu L,Zhou X,Cai W
Mon May 01 00:00:00 EDT 2023
Neuroprotective effects of a lead compound from coral via modulation of the orphan nuclear receptor Nurr1.
CNS neuroscience & therapeutics
Su JW,Yang P,Xing MM,Chen B,Xie XH,Ding JH,Lu M,Liu Y,Guo YW,Hu G
Wed Mar 01 00:00:00 EST 2023
Islet amyloid polypeptide triggers α-synuclein pathology in Parkinson's disease.
Progress in neurobiology
Meng L,Li Y,Liu C,Zhang G,Chen J,Xiong M,Pan L,Zhang X,Chen G,Xiong J,Liu C,Xu X,Bu L,Zhang Z,Zhang Z
Sat Jul 01 00:00:00 EDT 2023
Atmospheric particulate matter impairs cognition by modulating synaptic function via the nose-to-brain route.
The Science of the total environment
Liang C,Jiang Y,Zhang T,Ji Y,Zhang Y,Sun Y,Li S,Qi Y,Wang Y,Cai Y,Lai T,Cui L
Fri Jan 20 00:00:00 EST 2023
Choroid Plexus Modulates Subventricular Zone Adult Neurogenesis and Olfaction Through Secretion of Small Extracellular Vesicles.
bioRxiv : the preprint server for biology
Liu LL,Shannahan J,Zheng W
Sat Mar 18 00:00:00 EDT 2023
Neuro death through autophagy via the acetylation of FoxO1 by SIRT2 in the hippocampus of mice in a autism spectrum disorder mice model.
Journal of cellular physiology
Guo L,Jiang ZM,Zhan YJ,Pan W,Wu QW,Song FX,Zhou X,Zhou XY,Liu LJ,Wang JT
Thu Jun 01 00:00:00 EDT 2023
Neuro death through autophagy via the acetylation of FoxO1 by SIRT2 in the hippocampus of mice in a autism spectrum disorder mice model.
Journal of cellular physiology
Guo L,Jiang ZM,Zhan YJ,Pan W,Wu QW,Song FX,Zhou X,Zhou XY,Liu LJ,Wang JT
Thu Jun 01 00:00:00 EDT 2023
A versatile vessel casting method for fine mapping of vascular networks using a hydrogel-based lipophilic dye solution.
Cell reports methods
Zhu J,Liu X,Xu J,Deng Y,Wang P,Liu Z,Yang Q,Li D,Yu T,Zhu D
Mon Feb 27 00:00:00 EST 2023
Kctd7 deficiency induces myoclonic seizures associated with Purkinje cell death and microvascular defects.
Disease models & mechanisms
Liang JH,Alevy J,Akhanov V,Seo R,Massey CA,Jiang D,Zhou J,Sillitoe RV,Noebels JL,Samuel MA
Thu Sep 01 00:00:00 EDT 2022
Exercise-induced neuroprotection against cerebral ischemia/reperfusion injury is mediated via alleviating inflammasome-induced pyroptosis.
Experimental neurology
Liu MX,Luo L,Fu JH,He JY,Chen MY,He ZJ,Jia J
Tue Mar 01 00:00:00 EST 2022
A Class I HDAC Inhibitor BG45 Alleviates Cognitive Impairment through the CaMKII/ITPKA/Ca2+ Signaling Pathway.
Pharmaceuticals (Basel, Switzerland)
Liu J,Zhang C,Wang J,Huang Y,Shen D,Hu Y,Chu H,Yu X,Zhang L,Ma H
Mon Nov 28 00:00:00 EST 2022
miR-92a-2-5p Regulates the Proliferation and Differentiation of ASD-Derived Neural Progenitor Cells.
Current issues in molecular biology
Zhuang W,Liu H,He Z,Ju J,Gao Q,Shan Z,Lei L
Tue May 24 00:00:00 EDT 2022
Tubulin Isotypes and Posttranslational Modifications in Vascular Dementia and Alzheimer's Disease.
Journal of Alzheimer's disease reports
Santiago-Mujika E,Luthi-Carter R,Giorgini F,Mukaetova-Ladinska EB
Wed Jan 11 00:00:00 EST 2023
Tau accelerates α-synuclein aggregation and spreading in Parkinson's disease.
Brain : a journal of neurology
Pan L,Li C,Meng L,Tian Y,He M,Yuan X,Zhang G,Zhang Z,Xiong J,Chen G,Zhang Z
Fri Oct 21 00:00:00 EDT 2022
Tau accelerates α-synuclein aggregation and spreading in Parkinson's disease.
Brain : a journal of neurology
Pan L,Li C,Meng L,Tian Y,He M,Yuan X,Zhang G,Zhang Z,Xiong J,Chen G,Zhang Z
Fri Oct 21 00:00:00 EDT 2022
A functional SNP rs895819 on pre-miR-27a is associated with bipolar disorder by targeting NCAM1.
Communications biology
Yang Y,Lu W,Ning M,Zhou X,Wan X,Mi Q,Yang X,Zhang D,Zhang Y,Jiang B,He L,Liu J,Zou Y
Mon Apr 04 00:00:00 EDT 2022
Microglial NLRP3 inflammasome activates neurotoxic astrocytes in depression-like mice.
Cell reports
Li S,Fang Y,Zhang Y,Song M,Zhang X,Ding X,Yao H,Chen M,Sun Y,Ding J,Wang Q,Lu M,Wu G,Hu G
Tue Oct 25 00:00:00 EDT 2022
GPBAR1 preserves neurite and synapse of dopaminergic neurons via RAD21-OPCML signaling: Role in preventing Parkinson's disease in mouse model and human patients.
Pharmacological research
Zhang Y,Sun X,Zhang Y,Kang Z,Cai L,Ding J,Lu M,Hu G
Sat Oct 01 00:00:00 EDT 2022
Ectosomes and exosomes modulate neuronal spontaneous activity.
Journal of proteomics
Brás IC,Khani MH,Riedel D,Parfentev I,Gerhardt E,van Riesen C,Urlaub H,Gollisch T,Outeiro TF
Sun Oct 30 00:00:00 EDT 2022
DAPT Attenuates Cadmium-Induced Toxicity in Mice by Inhibiting Inflammation and the Notch/HES-1 Signaling Axis.
Frontiers in pharmacology
Yang JY,Shen DY,Wang J,Dai JF,Qin XY,Hu Y,Lan R
Sat Jul 16 00:00:00 EDT 2022
Salidroside alleviates cadmium-induced toxicity in mice by restoring the notch/HES-1 and RIP1-driven inflammatory signaling axis.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Shu XM,Hu Y,Fang X,Wang J,Qin XY,Lan R
Wed Jun 01 00:00:00 EDT 2022
Coeloglossum viride Var. Bracteatum Extract Attenuates MPTP-Induced Neurotoxicity in vivo by Restoring BDNF-TrkB and FGF2-Akt Signaling Axis and Inhibiting RIP1-Driven Inflammation.
Frontiers in pharmacology
Lang XY,Hu Y,Bai JP,Wang J,Qin XY,Lan R
Thu May 19 00:00:00 EDT 2022
Tandem mass tag-based quantitative proteomic analysis of effects of multiple sevoflurane exposures on the cerebral cortex of neonatal and adult mice.
Frontiers in neurology
Feng J,Lin H,Zhao Y,Yang Y,Zhuang X,Yu Y,Yu Y
Tue Jan 03 00:00:00 EST 2023
Pregabalin mitigates microglial activation and neuronal injury by inhibiting HMGB1 signaling pathway in radiation-induced brain injury.
Journal of neuroinflammation
Zhang Z,Jiang J,He Y,Cai J,Xie J,Wu M,Xing M,Zhang Z,Chang H,Yu P,Chen S,Yang Y,Shi Z,Liu Q,Sun H,He B,Zeng J,Huang J,Chen J,Li H,Li Y,Lin WJ,Tang Y
Wed Sep 21 00:00:00 EDT 2022
SKF83959 Attenuates Memory Impairment and Depressive-like Behavior during the Latent Period of Epilepsy via Allosteric Activation of the Sigma-1 Receptor.
ACS chemical neuroscience
Guo L,Gao T,Jia X,Gao C,Tian H,Wei Y,Lu W,Liu Z,Wang Y
Wed Nov 16 00:00:00 EST 2022
Recombinant human erythropoietin protects long-term cultured ageing primary nerve cells by upregulating the PI3K/Akt pathway.
Neuroreport
Wang H,Chen M,Gao Z,Gong Y,Yu X,Wu H
Wed Mar 02 00:00:00 EST 2022
Therapeutic Effect of Biomimetic Scaffold Loaded with Human Amniotic Epithelial Cell-Derived Neural-like Cells for Spinal Cord Injury.
Bioengineering (Basel, Switzerland)
Qiu C,Sun Y,Li J,Xu Y,Zhou J,Qiu C,Zhang S,He Y,Yu L
Sun Oct 09 00:00:00 EDT 2022
Human PMSCs-derived small extracellular vesicles alleviate neuropathic pain through miR-26a-5p/Wnt5a in SNI mice model.
Journal of neuroinflammation
Lu Y,Zhang J,Zeng F,Wang P,Guo X,Wang H,Qin Z,Tao T
Wed Sep 07 00:00:00 EDT 2022
A three-dimensional matrix system containing melatonin and neural stem cells repairs damage from traumatic brain injury in rats.
Neural regeneration research
Fang XY,Zhao DW,Zhang C,Ge HF,Zhang XY,Zhao FC,Jiang YB,Feng H,Hu R
Tue Nov 01 00:00:00 EDT 2022
The LPA-CDK5-tau pathway mediates neuronal injury in an in vitro model of ischemia-reperfusion insult.
BMC neurology
Wang Y,Zhang J,Huang L,Mo Y,Wang C,Li Y,Zhang Y,Zhang Z
Mon May 02 00:00:00 EDT 2022
Rare KCND3 Loss-of-Function Mutation Associated With the SCA19/22.
Frontiers in molecular neuroscience
Li M,Liu F,Hao X,Fan Y,Li J,Hu Z,Shi J,Fan L,Zhang S,Ma D,Guo M,Xu Y,Shi C
Sat Jul 16 00:00:00 EDT 2022
Coeloglossum viride var. bracteatum extract attenuates Aβ-induced toxicity by inhibiting RIP1-driven inflammation and necroptosis.
Journal of ethnopharmacology
Li XX,Lang XY,Ren TT,Wang J,Lan R,Qin XY
Mon Jan 10 00:00:00 EST 2022
Brain Microvascular Endothelial Cell-Derived Exosomes Protect Neurons from Ischemia-Reperfusion Injury in Mice.
Pharmaceuticals (Basel, Switzerland)
Sun J,Yuan Q,Guo L,Xiao G,Zhang T,Liang B,Yao R,Zhu Y,Li Y,Hu L
Wed Oct 19 00:00:00 EDT 2022
Brain Microvascular Endothelial Cell-Derived Exosomes Protect Neurons from Ischemia-Reperfusion Injury in Mice.
Pharmaceuticals (Basel, Switzerland)
Sun J,Yuan Q,Guo L,Xiao G,Zhang T,Liang B,Yao R,Zhu Y,Li Y,Hu L
Wed Oct 19 00:00:00 EDT 2022
Fine-tuning of mTOR signaling by the UBE4B-KLHL22 E3 ubiquitin ligase cascade in brain development.
Development (Cambridge, England)
Kong X,Shu X,Wang J,Liu D,Ni Y,Zhao W,Wang L,Gao Z,Chen J,Yang B,Guo X,Wang Z
Thu Dec 15 00:00:00 EST 2022
Electroacupuncture treatment improves motor function and neurological outcomes after cerebral ischemia/reperfusion injury.
Neural regeneration research
Li SS,Hua XY,Zheng MX,Wu JJ,Ma ZZ,Xing XX,Ma J,Shan CL,Xu JG
Fri Jul 01 00:00:00 EDT 2022
Electroacupuncture treatment improves motor function and neurological outcomes after cerebral ischemia/reperfusion injury.
Neural regeneration research
Li SS,Hua XY,Zheng MX,Wu JJ,Ma ZZ,Xing XX,Ma J,Shan CL,Xu JG
Fri Jul 01 00:00:00 EDT 2022
Ferroptosis participates in neuron damage in experimental cerebral malaria and is partially induced by activated CD8+ T cells.
Molecular brain
Liang J,Shen Y,Wang Y,Huang Y,Wang J,Zhu Q,Tong G,Yu K,Cao W,Wang Q,Li Y,Zhao Y
Mon Jun 20 00:00:00 EDT 2022
Exploring the pharmacological mechanism of calculus bovis in cerebral ischaemic stroke using a network pharmacology approach.
Journal of ethnopharmacology
Du X,Li C,Zhang S,Sun C,Zhang X,Chen C,Wang X,Cheng F,Wang Q
Thu Feb 10 00:00:00 EST 2022
Astragalin attenuates AlCl3/D-galactose-induced aging-like disorders by inhibiting oxidative stress and neuroinflammation.
Neurotoxicology
Hu Y,Fang X,Wang J,Ren TT,Zhao YY,Dai JF,Qin XY,Lan R
Fri Jul 01 00:00:00 EDT 2022
MPO/HOCl Facilitates Apoptosis and Ferroptosis in the SOD1G93A Motor Neuron of Amyotrophic Lateral Sclerosis.
Oxidative medicine and cellular longevity
Peng J,Pan J,Mo J,Peng Y
Fri Mar 18 00:00:00 EDT 2022
Islet amyloid polypeptide cross-seeds tau and drives the neurofibrillary pathology in Alzheimer's disease.
Molecular neurodegeneration
Zhang G,Meng L,Wang Z,Peng Q,Chen G,Xiong J,Zhang Z
Sat Jan 29 00:00:00 EST 2022
Galectin-9/Tim-3 pathway mediates dopaminergic neurodegeneration in MPTP-induced mouse model of Parkinson's disease.
Frontiers in molecular neuroscience
Peng Q,Zhang G,Guo X,Dai L,Xiong M,Zhang Z,Chen L,Zhang Z
Sat Dec 10 00:00:00 EST 2022
High-mobility Group Box 1 Contributes to Hypoxic-Ischemic Brain Damage by Facilitating Imbalance of Microglial Polarization through RAGE-PI3K/Akt Pathway in Neonatal Rats.
International journal of medical sciences
Sun Y,Zhu X,Zhu K,Yu J,Cheng L,Hei M
Thu Dec 15 00:00:00 EST 2022
ALS-associated KIF5A mutations abolish autoinhibition resulting in a toxic gain of function.
Cell reports
Baron DM,Fenton AR,Saez-Atienzar S,Giampetruzzi A,Sreeram A,Keagle PJ,Doocy VR,Smith NJ,Danielson EW,Andresano M,McCormack MC,Garcia J,Bercier V,Van Den Bosch L,Brent JR,Fallini C,Traynor BJ,Holzbaur ELF,Landers JE
Tue Apr 05 00:00:00 EDT 2022
PINK1 overexpression prevents forskolin-induced tau hyperphosphorylation and oxidative stress in a rat model of Alzheimer's disease.
Acta pharmacologica Sinica
Wang XJ,Qi L,Cheng YF,Ji XF,Chi TY,Liu P,Zou LB
Mon Aug 01 00:00:00 EDT 2022
Edaravone attenuates H2O2 or glutamate-induced toxicity in hippocampal neurons and improves AlCl3/D-galactose induced cognitive impairment in mice.
Neurotoxicology
Wu HT,Yu Y,Li XX,Lang XY,Gu RZ,Fan SR,Fang X,Bai JP,Lan R,Qin XY
Thu Jul 01 00:00:00 EDT 2021
Low-Intensity Ultrasound Causes Direct Excitation of Auditory Cortical Neurons.
Neural plasticity
Qi X,Lyu K,Meng L,Li C,Zhang H,Niu L,Lin Z,Zheng H,Tang J
Mon Dec 06 00:00:00 EST 2021
A five-m6A regulatory gene signature is a prognostic biomarker in lung adenocarcinoma patients.
Aging
Wu X,Sheng H,Wang L,Xia P,Wang Y,Yu L,Lv W,Hu J
Fri Mar 26 00:00:00 EDT 2021
Edaravone attenuates cadmium-induced toxicity by inhibiting oxidative stress and inflammation in ICR mice.
Neurotoxicology
Fan SR,Ren TT,Yun MY,Lan R,Qin XY
Wed Sep 01 00:00:00 EDT 2021
Defective biosynthesis of ascorbic acid in Sod1-deficient mice results in lethal damage to lung tissue.
Free radical biology & medicine
Homma T,Takeda Y,Nakano T,Akatsuka S,Kinoshita D,Kurahashi T,Saitoh S,Yamada KI,Miyata S,Asao H,Goto K,Watanabe T,Watanabe M,Toyokuni S,Fujii J
Fri Jan 01 00:00:00 EST 2021
Loss of mitochondrial transcription factor A in neural stem cells leads to immature brain development and triggers the activation of the integral stress response in vivo.
PloS one
Kuroda R,Tominaga K,Kasashima K,Kuroiwa K,Sakashita E,Hayakawa H,Kouki T,Ohno N,Kawai K,Endo H
Thu Nov 04 00:00:00 EDT 2021
Inhibition of miR-331-3p and miR-9-5p ameliorates Alzheimer's disease by enhancing autophagy.
Theranostics
Chen ML,Hong CG,Yue T,Li HM,Duan R,Hu WB,Cao J,Wang ZX,Chen CY,Hu XK,Wu B,Liu HM,Tan YJ,Liu JH,Luo ZW,Zhang Y,Rao SS,Luo MJ,Yin H,Wang YY,Xia K,Tang SY,Xie H,Liu ZZ
Mon Aug 16 00:00:00 EDT 2021
Enriched environment remedies cognitive dysfunctions and synaptic plasticity through NMDAR-Ca2+-Activin A circuit in chronic cerebral hypoperfusion rats.
Aging
Zhang X,Shi X,Wang J,Xu Z,He J
Mon Aug 30 00:00:00 EDT 2021
Activation of ephrinb1/EPHB2/MAP-2/NMDAR Mediates Hippocampal Neurogenesis Promoted by Transcranial Direct Current Stimulation in Cerebral-Ischemic Mice.
Neuromolecular medicine
Ma X,Cheng O,Jiang Q,Yang J,Xiao H,Qiu H
Wed Dec 01 00:00:00 EST 2021
3D printable Sodium alginate-Matrigel (SA-MA) hydrogel facilitated ectomesenchymal stem cells (EMSCs) neuron differentiation.
Journal of biomaterials applications
Li Y,Cao X,Deng W,Yu Q,Sun C,Ma P,Shao F,Yusif MM,Ge Z,Wang K,Li R,Yu J,Xu X
Fri Jan 01 00:00:00 EST 2021
Vezatin regulates seizures by controlling AMPAR-mediated synaptic activity.
Cell death & disease
Wang Y,Yuan J,Yu X,Liu X,Tan C,Chen Y,Xu T
Tue Oct 12 00:00:00 EDT 2021
Astrocytic ApoE reprograms neuronal cholesterol metabolism and histone-acetylation-mediated memory.
Neuron
Li X,Zhang J,Li D,He C,He K,Xue T,Wan L,Zhang C,Liu Q
Wed Mar 17 00:00:00 EDT 2021
Astrocytic ApoE reprograms neuronal cholesterol metabolism and histone-acetylation-mediated memory.
Neuron
Li X,Zhang J,Li D,He C,He K,Xue T,Wan L,Zhang C,Liu Q
Wed Mar 17 00:00:00 EDT 2021
Gisenoside Rg1 attenuates cadmium-induced neurotoxicity in vitro and in vivo by attenuating oxidative stress and inflammation.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Ren TT,Yang JY,Wang J,Fan SR,Lan R,Qin XY
Wed Dec 01 00:00:00 EST 2021
Gisenoside Rg1 attenuates cadmium-induced neurotoxicity in vitro and in vivo by attenuating oxidative stress and inflammation.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Ren TT,Yang JY,Wang J,Fan SR,Lan R,Qin XY
Wed Dec 01 00:00:00 EST 2021
Upregulation of antioxidant and autophagy pathways via NRF2 activation protects spinal cord neurons from ozone damage.
Molecular medicine reports
Zhang C,Ma S,Zhao X,Wen B,Sun P,Fu Z
Tue Jun 01 00:00:00 EDT 2021
Cardiac glycosaminoglycans and structural alterations during chronic stress-induced depression-like behavior in mice.
American journal of physiology. Heart and circulatory physiology
Luong H,Singh S,Patil M,Krishnamurthy P
Sat May 01 00:00:00 EDT 2021
Transgenic Mice Expressing Human α-Synuclein 1-103 Fragment as a Novel Model of Parkinson's Disease.
Frontiers in aging neuroscience
Tian Y,He M,Pan L,Yuan X,Xiong M,Meng L,Yao Z,Yu Z,Ye K,Zhang Z
Wed Nov 10 00:00:00 EST 2021
EGB761 ameliorates chronic cerebral hypoperfusion-induced cognitive dysfunction and synaptic plasticity impairment.
Aging
Yao ZH,Wang J,Yuan JP,Xiao K,Zhang SF,Xie YC,Mei JH
Wed Feb 03 00:00:00 EST 2021
EGB761 ameliorates chronic cerebral hypoperfusion-induced cognitive dysfunction and synaptic plasticity impairment.
Aging
Yao ZH,Wang J,Yuan JP,Xiao K,Zhang SF,Xie YC,Mei JH
Wed Feb 03 00:00:00 EST 2021
In vivo reprogramming of NG2 glia enables adult neurogenesis and functional recovery following spinal cord injury.
Cell stem cell
Tai W,Wu W,Wang LL,Ni H,Chen C,Yang J,Zang T,Zou Y,Xu XM,Zhang CL
Thu May 06 00:00:00 EDT 2021
Maternal high sugar and fat diet benefits offspring brain function via targeting on the gut-brain axis.
Aging
Wang D,Zhang H,Zeng M,Tang X,Zhu X,Guo Y,Qi L,Xie Y,Zhang M,Chen D
Fri Mar 26 00:00:00 EDT 2021
Thymosin β4 reverses phenotypic polarization of glial cells and cognitive impairment via negative regulation of NF-κB signaling axis in APP/PS1 mice.
Journal of neuroinflammation
Wang M,Feng LR,Li ZL,Ma KG,Chang KW,Chen XL,Yang PB,Ji SF,Ma YB,Han H,Ruganzua JB,Yang WN,Qian YH
Mon Jun 28 00:00:00 EDT 2021
Thymosin β4 reverses phenotypic polarization of glial cells and cognitive impairment via negative regulation of NF-κB signaling axis in APP/PS1 mice.
Journal of neuroinflammation
Wang M,Feng LR,Li ZL,Ma KG,Chang KW,Chen XL,Yang PB,Ji SF,Ma YB,Han H,Ruganzua JB,Yang WN,Qian YH
Mon Jun 28 00:00:00 EDT 2021
Protection of hUC-MSCs against neuronal complement C3a receptor-mediated NLRP3 activation in CUMS-induced mice.
Neuroscience letters
Li J,Tian S,Wang H,Wang Y,Du C,Fang J,Wang X,Wang Y,Gong Z,Yan B,Wang M
Sun Jan 10 00:00:00 EST 2021
Resveratrol-mediated neurorestoration after cerebral ischemic injury - Sonic Hedgehog signaling pathway.
Life sciences
Yu P,Wang L,Tang F,Guo S,Liao H,Fan C,Yang Q
Wed Sep 01 00:00:00 EDT 2021
Maternal Ethanol Exposure Acutely Elevates Src Family Kinase Activity in the Fetal Cortex.
Molecular neurobiology
Wang D,Howell BW,Olson EC
Fri Oct 01 00:00:00 EDT 2021
Rhein Relieves Oxidative Stress in an Aβ1-42 Oligomer-Burdened Neuron Model by Activating the SIRT1/PGC-1α-Regulated Mitochondrial Biogenesis.
Frontiers in pharmacology
Yin Z,Geng X,Zhang Z,Wang Y,Gao X
Wed Sep 29 00:00:00 EDT 2021
Quercetin hinders microglial activation to alleviate neurotoxicity via the interplay between NLRP3 inflammasome and mitophagy.
Redox biology
Han X,Xu T,Fang Q,Zhang H,Yue L,Hu G,Sun L
Sun Aug 01 00:00:00 EDT 2021
Silencing of circIgf1r plays a protective role in neuronal injury via regulating astrocyte polarization during epilepsy.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Shao L,Jiang GT,Yang XL,Zeng ML,Cheng JJ,Kong S,Dong X,Chen TX,Han S,Yin J,Liu WH,He XH,He C,Peng BW
Mon Feb 01 00:00:00 EST 2021
Photobiomodulation Therapy Ameliorates Glutamatergic Dysfunction in Mice with Chronic Unpredictable Mild Stress-Induced Depression.
Oxidative medicine and cellular longevity
Zhang D,Shen Q,Wu X,Xing D
Wed Jun 09 00:00:00 EDT 2021
Reactive microglia enhance the transmission of exosomal α-synuclein via toll-like receptor 2.
Brain : a journal of neurology
Xia Y,Zhang G,Kou L,Yin S,Han C,Hu J,Wan F,Sun Y,Wu J,Li Y,Huang J,Xiong N,Zhang Z,Wang T
Tue Aug 17 00:00:00 EDT 2021
The Non-hormonal Male Contraceptive Adjudin Exerts its Effects via MAPs and Signaling Proteins mTORC1/rpS6 and FAK-Y407.
Endocrinology
Wang L,Yan M,Li H,Wu S,Ge R,Wong CKC,Silvestrini B,Sun F,Cheng CY
Fri Jan 01 00:00:00 EST 2021
The Role of HSP90α in Methamphetamine/Hyperthermia-Induced Necroptosis in Rat Striatal Neurons.
Frontiers in pharmacology
Liao LS,Lu S,Yan WT,Wang SC,Guo LM,Yang YD,Huang K,Hu XM,Zhang Q,Yan J,Xiong K
Fri Aug 06 00:00:00 EDT 2021
Kir6.2 is essential to maintain neurite features by modulating PM20D1-reduced mitochondrial ATP generation.
Redox biology
Song N,Fang Y,Zhu H,Liu J,Jiang S,Sun S,Xu R,Ding J,Hu G,Lu M
Mon Nov 01 00:00:00 EDT 2021
Kir6.2 is essential to maintain neurite features by modulating PM20D1-reduced mitochondrial ATP generation.
Redox biology
Song N,Fang Y,Zhu H,Liu J,Jiang S,Sun S,Xu R,Ding J,Hu G,Lu M
Mon Nov 01 00:00:00 EDT 2021
Remote Limb Ischemic Postconditioning Protects Against Ischemic Stroke by Promoting Regulatory T Cells Thriving.
Journal of the American Heart Association
Yu HH,Ma XT,Ma X,Chen M,Chu YH,Wu LJ,Wang W,Qin C,Tian DS
Tue Nov 16 00:00:00 EST 2021
Effects of the histone acetylase inhibitor C646 on growth and differentiation of adipose-derived stem cells.
Cell cycle (Georgetown, Tex.)
Wang Z,Wang X,Bi M,Hu X,Wang Q,Liang H,Liu D
Mon Feb 01 00:00:00 EST 2021
XQ-1H promotes cerebral angiogenesis via activating PI3K/Akt/GSK3β/β-catenin/VEGF signal in mice exposed to cerebral ischemic injury.
Life sciences
Fei Y,Zhao B,Zhu J,Fang W,Li Y
Sat May 01 00:00:00 EDT 2021
Complement C3 Aggravates Post-epileptic Neuronal Injury Via Activation of TRPV1.
Neuroscience bulletin
Jiang GT,Shao L,Kong S,Zeng ML,Cheng JJ,Chen TX,Han S,Yin J,Liu WH,He XH,Liu YM,Gongga L,Peng BW
Fri Oct 01 00:00:00 EDT 2021
SARS-CoV-2 targets neurons of 3D human brain organoids.
The EMBO journal
Ramani A,Müller L,Ostermann PN,Gabriel E,Abida-Islam P,Müller-Schiffmann A,Mariappan A,Goureau O,Gruell H,Walker A,Andrée M,Hauka S,Houwaart T,Dilthey A,Wohlgemuth K,Omran H,Klein F,Wieczorek D,Adams O,Timm J,Korth C,Schaal H,Gopalakrishnan J
Thu Oct 15 00:00:00 EDT 2020
Neuron-Derived Estrogen Is Critical for Astrocyte Activation and Neuroprotection of the Ischemic Brain.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Lu Y,Sareddy GR,Wang J,Zhang Q,Tang FL,Pratap UP,Tekmal RR,Vadlamudi RK,Brann DW
Wed Sep 16 00:00:00 EDT 2020
Small molecular compounds efficiently convert human fibroblasts directly into neurons.
Molecular medicine reports
Yang J,Cao H,Guo S,Zhu H,Tao H,Zhang L,Chen Z,Sun T,Chi S,Hu Q
Tue Dec 01 00:00:00 EST 2020
Evidence for the spread of human-derived mutant huntingtin protein in mice and non-human primates.
Neurobiology of disease
Gosset P,Maxan A,Alpaugh M,Breger L,Dehay B,Tao Z,Ling Z,Qin C,Cisbani G,Fortin N,Vonsattel JG,Lacroix S,Oueslati A,Bezard E,Cicchetti F
Wed Jul 01 00:00:00 EDT 2020
Rapid and Efficient Invasion Assay of Glioblastoma in Human Brain Organoids.
Cell reports
Goranci-Buzhala G,Mariappan A,Gabriel E,Ramani A,Ricci-Vitiani L,Buccarelli M,D'Alessandris QG,Pallini R,Gopalakrishnan J
Tue Jun 09 00:00:00 EDT 2020
Dose-dependent effects of tetramethylpyrazine on the characteristics of human umbilical cord mesenchymal stem cells for stroke therapy.
Neuroscience letters
Cao H,Zhu X,Zhang J,Xu M,Ge L,Zhang C
Mon Mar 23 00:00:00 EDT 2020
Dose-dependent effects of tetramethylpyrazine on the characteristics of human umbilical cord mesenchymal stem cells for stroke therapy.
Neuroscience letters
Cao H,Zhu X,Zhang J,Xu M,Ge L,Zhang C
Mon Mar 23 00:00:00 EDT 2020
NLRP3-dependent pyroptosis is required for HIV-1 gp120-induced neuropathology.
Cellular & molecular immunology
He X,Yang W,Zeng Z,Wei Y,Gao J,Zhang B,Li L,Liu L,Wan Y,Zeng Q,Gong Z,Liu L,Zhang H,Li Y,Yang S,Hu T,Wu L,Masliah E,Huang S,Cao H
Sun Mar 01 00:00:00 EST 2020
NLRP3-dependent pyroptosis is required for HIV-1 gp120-induced neuropathology.
Cellular & molecular immunology
He X,Yang W,Zeng Z,Wei Y,Gao J,Zhang B,Li L,Liu L,Wan Y,Zeng Q,Gong Z,Liu L,Zhang H,Li Y,Yang S,Hu T,Wu L,Masliah E,Huang S,Cao H
Sun Mar 01 00:00:00 EST 2020
Qingxin Kaiqiao Fang Inhibits Aβ 25-35-Induced Apoptosis in Primary Cultured Rat Hippocampal Neuronal Cells via the p38 MAPK Pathway: An Experimental Validation and Network Pharmacology Study.
Evidence-based complementary and alternative medicine : eCAM
Wang TQ,Lai XX,Xu LT,Shen Y,Lin JW,Gao SY,Hu HY
Mon Sep 28 00:00:00 EDT 2020
Actin Alpha 2 (ACTA2) Downregulation Inhibits Neural Stem Cell Migration through Rho GTPase Activation.
Stem cells international
Zhang J,Jiang X,Zhang C,Zhong J,Fang X,Li H,Xie F,Huang X,Zhang X,Hu Q,Ge H,Yu A
Mon Sep 28 00:00:00 EDT 2020
Generation of an iPSC line (HUSTi002-A) from fibroblasts of a patient with Sertoli cell-only syndrome carrying c.731_732delAT in PIWIL2 gene.
Stem cell research
Wang X,Xie S,Li Z,Ye Z,Gu X,Zhou L,Li H
Wed Jan 01 00:00:00 EST 2020
Ablation of GSDMD Improves Outcome of Ischemic Stroke Through Blocking Canonical and Non-canonical Inflammasomes Dependent Pyroptosis in Microglia.
Frontiers in neurology
Wang K,Sun Z,Ru J,Wang S,Huang L,Ruan L,Lin X,Jin K,Zhuge Q,Yang S
Fri Dec 18 00:00:00 EST 2020
Bisphenol-A exposure induced neurotoxicity and associated with synapse and cytoskeleton in Neuro-2a cells.
Toxicology in vitro : an international journal published in association with BIBRA
Yin Z,Hua L,Chen L,Hu D,Li J,An Z,Tian T,Ning H,Ge Y
Tue Sep 01 00:00:00 EDT 2020
Bisphenol-A exposure induced neurotoxicity and associated with synapse and cytoskeleton in Neuro-2a cells.
Toxicology in vitro : an international journal published in association with BIBRA
Yin Z,Hua L,Chen L,Hu D,Li J,An Z,Tian T,Ning H,Ge Y
Tue Sep 01 00:00:00 EDT 2020
Single-Cell Analysis for Glycogen Localization and Metabolism in Cultured Astrocytes.
Cellular and molecular neurobiology
Zhu Y,Fan Z,Wang R,Xie R,Guo H,Zhang M,Guo B,Sun T,Zhang H,Zhuo L,Li Y,Wu S
Wed Jul 01 00:00:00 EDT 2020
Single-Cell Analysis for Glycogen Localization and Metabolism in Cultured Astrocytes.
Cellular and molecular neurobiology
Zhu Y,Fan Z,Wang R,Xie R,Guo H,Zhang M,Guo B,Sun T,Zhang H,Zhuo L,Li Y,Wu S
Wed Jul 01 00:00:00 EDT 2020
MicroRNA Let-7b-5p Induces Electroacupuncture Tolerance by Downregulating the MKP-1 Gene in Rats Subjected to CFA-induced Inflammatory Nociception.
Journal of molecular neuroscience : MN
Zhang Q,Abouelfetouh MM,Chen S,Li M,Ding M,Ding Y
Sat Aug 01 00:00:00 EDT 2020
5-HT1AR alleviates Aβ-induced cognitive decline and neuroinflammation through crosstalk with NF-κB pathway in mice.
International immunopharmacology
Wang M,Zong HF,Chang KW,Han H,Yasir Rizvi M,Iffat Neha S,Li ZY,Yang WN,Qian YH
Tue Mar 03 00:00:00 EST 2020
Therapeutic mechanism of human neural stem cell-derived extracellular vesicles against hypoxia-reperfusion injury in vitro.
Life sciences
Liu Q,Tan Y,Qu T,Zhang J,Duan X,Xu H,Mu Y,Ma H,Wang F
Sat Aug 01 00:00:00 EDT 2020
Plasmacytoid Dendritic Cells Protect Against Middle Cerebral Artery Occlusion Induced Brain Injury by Priming Regulatory T Cells.
Frontiers in cellular neuroscience
Chen C,Chencheng Z,Cuiying L,Xiaokun G
Mon Sep 28 00:00:00 EDT 2020
Cofilin 1 promotes the aggregation and cell-to-cell transmission of α-synuclein in Parkinson's disease.
Biochemical and biophysical research communications
Yan M,Meng L,Dai L,Zhang X,Chen G,Zheng Y,Zha Y,Zeng Y,Zhang Z
Thu Sep 03 00:00:00 EDT 2020
RNA Binding Protein Motif 3 Inhibits Oxygen-Glucose Deprivation/Reoxygenation-Induced Apoptosis Through Promoting Stress Granules Formation in PC12 Cells and Rat Primary Cortical Neurons.
Frontiers in cellular neuroscience
Si W,Li Z,Huang Z,Ye S,Li X,Li Y,Kuang W,Chen D,Zhu M
Thu Oct 01 00:00:00 EDT 2020
Premature termination codon readthrough upregulates progranulin expression and improves lysosomal function in preclinical models of GRN deficiency.
Molecular neurodegeneration
Frew J,Baradaran-Heravi A,Balgi AD,Wu X,Yan TD,Arns S,Shidmoossavee FS,Tan J,Jaquith JB,Jansen-West KR,Lynn FC,Gao FB,Petrucelli L,Feldman HH,Mackenzie IR,Roberge M,Nygaard HB
Mon Mar 16 00:00:00 EDT 2020
Alix and Syntenin-1 direct amyloid precursor protein trafficking into extracellular vesicles.
BMC molecular and cell biology
Cone AS,Hurwitz SN,Lee GS,Yuan X,Zhou Y,Li Y,Meckes DG
Thu Jul 30 00:00:00 EDT 2020
GABA-mediated activated microglia induce neuroinflammation in the hippocampus of mice following cold exposure through the NLRP3 inflammasome and NF-κB signaling pathways.
International immunopharmacology
Lang L,Xu B,Yuan J,Li S,Lian S,Chen Y,Guo J,Yang H
Tue Dec 01 00:00:00 EST 2020
Methamphetamine induces GSDME-dependent cell death in hippocampal neuronal cells through the endoplasmic reticulum stress pathway.
Brain research bulletin
Liu Y,Wen D,Gao J,Xie B,Yu H,Shen Q,Zhang J,Jing W,Cong B,Ma C
Tue Sep 01 00:00:00 EDT 2020
Memantine can relieve the neuronal impairment caused by neurotropic virus infection.
Journal of medical virology
Sun L,Zhou M,Liu C,Tang Y,Xiao K,Dai J,Gao Z,Siew L,Cao G,Wu X,Li L,Zhang R
Sat Jun 01 00:00:00 EDT 2019
Memantine can relieve the neuronal impairment caused by neurotropic virus infection.
Journal of medical virology
Sun L,Zhou M,Liu C,Tang Y,Xiao K,Dai J,Gao Z,Siew L,Cao G,Wu X,Li L,Zhang R
Sat Jun 01 00:00:00 EDT 2019
A single factor induces neuronal differentiation to suppress glioma cell growth.
CNS neuroscience & therapeutics
Fu JQ,Chen Z,Hu YJ,Fan ZH,Guo ZX,Liang JY,Ryu BM,Ren JL,Shi XJ,Li J,Jia S,Wang J,Ke XS,Ma X,Tan X,Zhang T,Chen XZ,Zhang C
Mon Apr 01 00:00:00 EDT 2019
A single factor induces neuronal differentiation to suppress glioma cell growth.
CNS neuroscience & therapeutics
Fu JQ,Chen Z,Hu YJ,Fan ZH,Guo ZX,Liang JY,Ryu BM,Ren JL,Shi XJ,Li J,Jia S,Wang J,Ke XS,Ma X,Tan X,Zhang T,Chen XZ,Zhang C
Mon Apr 01 00:00:00 EDT 2019
Treadmill exercise ameliorates focal cerebral ischemia/reperfusion-induced neurological deficit by promoting dendritic modification and synaptic plasticity via upregulating caveolin-1/VEGF signaling pathways.
Experimental neurology
Xie Q,Cheng J,Pan G,Wu S,Hu Q,Jiang H,Wang Y,Xiong J,Pang Q,Chen X
Fri Mar 01 00:00:00 EST 2019
Treadmill exercise ameliorates focal cerebral ischemia/reperfusion-induced neurological deficit by promoting dendritic modification and synaptic plasticity via upregulating caveolin-1/VEGF signaling pathways.
Experimental neurology
Xie Q,Cheng J,Pan G,Wu S,Hu Q,Jiang H,Wang Y,Xiong J,Pang Q,Chen X
Fri Mar 01 00:00:00 EST 2019
Treadmill exercise ameliorates focal cerebral ischemia/reperfusion-induced neurological deficit by promoting dendritic modification and synaptic plasticity via upregulating caveolin-1/VEGF signaling pathways.
Experimental neurology
Xie Q,Cheng J,Pan G,Wu S,Hu Q,Jiang H,Wang Y,Xiong J,Pang Q,Chen X
Fri Mar 01 00:00:00 EST 2019
Treadmill exercise ameliorates focal cerebral ischemia/reperfusion-induced neurological deficit by promoting dendritic modification and synaptic plasticity via upregulating caveolin-1/VEGF signaling pathways.
Experimental neurology
Xie Q,Cheng J,Pan G,Wu S,Hu Q,Jiang H,Wang Y,Xiong J,Pang Q,Chen X
Fri Mar 01 00:00:00 EST 2019
Deletion of Tbk1 disrupts autophagy and reproduces behavioral and locomotor symptoms of FTD-ALS in mice.
Aging
Duan W,Guo M,Yi L,Zhang J,Bi Y,Liu Y,Li Y,Li Z,Ma Y,Zhang G,Liu Y,Song X,Li C
Tue Apr 30 00:00:00 EDT 2019
Deletion of Tbk1 disrupts autophagy and reproduces behavioral and locomotor symptoms of FTD-ALS in mice.
Aging
Duan W,Guo M,Yi L,Zhang J,Bi Y,Liu Y,Li Y,Li Z,Ma Y,Zhang G,Liu Y,Song X,Li C
Tue Apr 30 00:00:00 EDT 2019
Deletion of Tbk1 disrupts autophagy and reproduces behavioral and locomotor symptoms of FTD-ALS in mice.
Aging
Duan W,Guo M,Yi L,Zhang J,Bi Y,Liu Y,Li Y,Li Z,Ma Y,Zhang G,Liu Y,Song X,Li C
Tue Apr 30 00:00:00 EDT 2019
Zn2+ Aggravates Tau Aggregation and Neurotoxicity.
International journal of molecular sciences
Li X,Du X,Ni J
Wed Jan 23 00:00:00 EST 2019
Cortisol Excess-Mediated Mitochondrial Damage Induced Hippocampal Neuronal Apoptosis in Mice Following Cold Exposure.
Cells
Xu B,Lang LM,Li SZ,Guo JR,Wang JF,Wang D,Zhang LP,Yang HM,Lian S
Tue Jun 18 00:00:00 EDT 2019
Inhibitory effects of glucagon-like peptide-1 receptor on epilepsy.
Biochemical and biophysical research communications
Wen Y,Wu K,Xie Y,Dan W,Zhan Y,Shi Q
Tue Mar 26 00:00:00 EDT 2019
Microglia as modulators of exosomal alpha-synuclein transmission.
Cell death & disease
Xia Y,Zhang G,Han C,Ma K,Guo X,Wan F,Kou L,Yin S,Liu L,Huang J,Xiong N,Wang T
Wed Feb 20 00:00:00 EST 2019
Toxicity mechanism of sevoflurane in neural stem cells of rats through DNA methylation.
Experimental and therapeutic medicine
Wang K,Tian Y,Zhang Y,Li X,Wei X,Hu H,Xu S
Mon Jul 01 00:00:00 EDT 2019
Hyperbaric oxygen therapy reduces apoptosis and dendritic/synaptic degeneration via the BDNF/TrkB signaling pathways in SCI rats.
Life sciences
Ying X,Tu W,Li S,Wu Q,Chen X,Zhou Y,Hu J,Yang G,Jiang S
Mon Jul 15 00:00:00 EDT 2019
Hyperbaric oxygen therapy reduces apoptosis and dendritic/synaptic degeneration via the BDNF/TrkB signaling pathways in SCI rats.
Life sciences
Ying X,Tu W,Li S,Wu Q,Chen X,Zhou Y,Hu J,Yang G,Jiang S
Mon Jul 15 00:00:00 EDT 2019
Probucol, a "non-statin" cholesterol-lowering drug, ameliorates D-galactose induced cognitive deficits by alleviating oxidative stress via Keap1/Nrf2 signaling pathway in mice.
Aging
Huang JL,Yu C,Su M,Yang SM,Zhang F,Chen YY,Liu JY,Jiang YF,Zhong ZG,Wu DP
Mon Oct 07 00:00:00 EDT 2019
Lentiviral-mediated ephrin B2 gene modification of rat bone marrow mesenchymal stem cells.
The Journal of international medical research
Zhu M,Hua Y,Tang J,Zhao X,Zhang L,Zhang Y
Mon Jul 01 00:00:00 EDT 2019
Talin promotes integrin activation accompanied by generation of tension in talin and an increase in osmotic pressure in neurite outgrowth.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wang Y,Zhang X,Tian J,Shan J,Hu Y,Zhai Y,Guo J
Wed May 01 00:00:00 EDT 2019
Multi-site dynamic recording for Aβ oligomers-induced Alzheimer's disease in vitro based on neuronal network chip.
Biosensors & bioelectronics
Gao F,Gao K,He C,Liu M,Wan H,Wang P
Wed May 15 00:00:00 EDT 2019
JC virus infection of meningeal and choroid plexus cells in patients with progressive multifocal leukoencephalopathy.
Journal of neurovirology
Corbridge SM,Rice RC,Bean LA,Wüthrich C,Dang X,Nicholson DA,Koralnik IJ
Thu Aug 01 00:00:00 EDT 2019
Silencing Huwe1 reduces apoptosis of cortical neurons exposed to oxygen-glucose deprivation and reperfusion.
Neural regeneration research
He GQ,Xu WM,Liao HJ,Jiang C,Li CQ,Zhang W
Fri Nov 01 00:00:00 EDT 2019
Down-regulation of ATF1 leads to early neuroectoderm differentiation of human embryonic stem cells by increasing the expression level of SOX2.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Yang SC,Liu JJ,Wang CK,Lin YT,Tsai SY,Chen WJ,Huang WK,Tu PA,Lin YC,Chang CF,Cheng CL,Lin H,Lai CY,Lin CY,Lee YH,Chiu YC,Hsu CC,Hsu SC,Hsiao M,Schuyler SC,Lu FL,Lu J
Sun Sep 01 00:00:00 EDT 2019
High-Mobility Group Box 1 Contributes to Cerebral Cortex Injury in a Neonatal Hypoxic-Ischemic Rat Model by Regulating the Phenotypic Polarization of Microglia.
Frontiers in cellular neuroscience
Sun Y,Hei M,Fang Z,Tang Z,Wang B,Hu N
Wed Sep 30 00:00:00 EDT 2020
A dual-enzymatically cross-linked injectable gelatin hydrogel loaded with BMSC improves neurological function recovery of traumatic brain injury in rats.
Biomaterials science
Yao M,Gao F,Xu R,Zhang J,Chen Y,Guan F
Tue Sep 24 00:00:00 EDT 2019
Restorative effect of modified dioscorea pills on the structure of hippocampal neurovascular unit in an animal model of chronic cerebral hypoperfusion.
Heliyon
Li HB
Mon Apr 01 00:00:00 EDT 2019
Oxidation Stress-Mediated MAPK Signaling Pathway Activation Induces Neuronal Loss in the CA1 and CA3 Regions of the Hippocampus of Mice Following Chronic Cold Exposure.
Brain sciences
Xu B,Lang LM,Lian S,Guo JR,Wang JF,Yang HM,Li SZ
Sat Oct 12 00:00:00 EDT 2019
Oxidation Stress-Mediated MAPK Signaling Pathway Activation Induces Neuronal Loss in the CA1 and CA3 Regions of the Hippocampus of Mice Following Chronic Cold Exposure.
Brain sciences
Xu B,Lang LM,Lian S,Guo JR,Wang JF,Yang HM,Li SZ
Sat Oct 12 00:00:00 EDT 2019
Chondroitin sulfate proteoglycan represses neural stem/progenitor cells migration via PTPσ/α-actinin4 signaling pathway.
Journal of cellular biochemistry
Zhong J,Lan C,Zhang C,Yang Y,Chen WX,Zhang KY,Zhao HL,Fang XY,Li HH,Tan L,Wang P,Ge HF,Hu R,Feng H
Mon Jul 01 00:00:00 EDT 2019
Chondroitin sulfate proteoglycan represses neural stem/progenitor cells migration via PTPσ/α-actinin4 signaling pathway.
Journal of cellular biochemistry
Zhong J,Lan C,Zhang C,Yang Y,Chen WX,Zhang KY,Zhao HL,Fang XY,Li HH,Tan L,Wang P,Ge HF,Hu R,Feng H
Mon Jul 01 00:00:00 EDT 2019
Neuroprotective effects of a Smoothened receptor agonist against postoperative cognitive dysfunction by promoting autophagy in the dentate gyrus of aged rats.
Neurological research
Li PJ,Guo YQ,Ding PY,Liu RB,Deng F,Feng XX,Yan WJ
Tue Oct 01 00:00:00 EDT 2019
Microglia Activated by Excess Cortisol Induce HMGB1 Acetylation and Neuroinflammation in the Hippocampal DG Region of Mice Following Cold Exposure.
Biomolecules
Xu B,Lang LM,Li SZ,Guo JR,Wang JF,Yang HM,Lian S
Fri Aug 30 00:00:00 EDT 2019
Geniposide ameliorated fluoxetine-suppressed neurite outgrowth in Neuro2a neuroblastoma cells.
Life sciences
Chen MK,Peng CC,Maner RS,Zulkefli ND,Huang SM,Hsieh CL
Sat Jun 01 00:00:00 EDT 2019
Geniposide ameliorated fluoxetine-suppressed neurite outgrowth in Neuro2a neuroblastoma cells.
Life sciences
Chen MK,Peng CC,Maner RS,Zulkefli ND,Huang SM,Hsieh CL
Sat Jun 01 00:00:00 EDT 2019
Protective effect of 4-Methoxy benzyl alcohol on the neurovascular unit after cerebral ischemia reperfusion injury.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
He F,Dai R,Zhou X,Li X,Song X,Yan H,Meng Q,Yang C,Lin Q
Tue Oct 01 00:00:00 EDT 2019
Cysteinyl Leukotriene Receptor 2 is Involved in Inflammation and Neuronal Damage by Mediating Microglia M1/M2 Polarization through NF-κB Pathway.
Neuroscience
Zhao R,Ying M,Gu S,Yin W,Li Y,Yuan H,Fang S,Li M
Sun Dec 01 00:00:00 EST 2019
Outcome of cell suspension allografts in a patient with Huntington's disease.
Annals of neurology
Maxan A,Mason S,Saint-Pierre M,Smith E,Ho A,Harrower T,Watts C,Tai Y,Pavese N,Savage JC,Tremblay MÈ,Gould P,Rosser AE,Dunnett SB,Piccini P,Barker RA,Cicchetti F
Sat Dec 01 00:00:00 EST 2018
Outcome of cell suspension allografts in a patient with Huntington's disease.
Annals of neurology
Maxan A,Mason S,Saint-Pierre M,Smith E,Ho A,Harrower T,Watts C,Tai Y,Pavese N,Savage JC,Tremblay MÈ,Gould P,Rosser AE,Dunnett SB,Piccini P,Barker RA,Cicchetti F
Sat Dec 01 00:00:00 EST 2018
Tert-butylhydroquinone post-treatment attenuates neonatal hypoxic-ischemic brain damage in rats.
Neurochemistry international
Zhang J,Tucker LD,Lu Y,Yang L,Wu C,Li Y,Zhang Q
Fri Jun 01 00:00:00 EDT 2018
Tert-butylhydroquinone post-treatment attenuates neonatal hypoxic-ischemic brain damage in rats.
Neurochemistry international
Zhang J,Tucker LD,Lu Y,Yang L,Wu C,Li Y,Zhang Q
Fri Jun 01 00:00:00 EDT 2018
Perineurial-like Cells and EMA Expression in the Suprachoroidal Region of the Human Eye.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Gilbert AR,Chévez-Barrios P,Cykowski MD
Tue May 01 00:00:00 EDT 2018
Perineurial-like Cells and EMA Expression in the Suprachoroidal Region of the Human Eye.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Gilbert AR,Chévez-Barrios P,Cykowski MD
Tue May 01 00:00:00 EDT 2018
Characterization of 3D embryonic C57BL/6 and A/J mouse midbrain micromass in vitro culture systems for developmental neurotoxicity testing.
Toxicology in vitro : an international journal published in association with BIBRA
Park JJ,Weldon BA,Hong S,Workman T,Griffith WC,Park JH,Faustman EM
Sun Apr 01 00:00:00 EDT 2018
Characterization of 3D embryonic C57BL/6 and A/J mouse midbrain micromass in vitro culture systems for developmental neurotoxicity testing.
Toxicology in vitro : an international journal published in association with BIBRA
Park JJ,Weldon BA,Hong S,Workman T,Griffith WC,Park JH,Faustman EM
Sun Apr 01 00:00:00 EDT 2018
Neuroserpin restores autophagy and promotes functional recovery after acute spinal cord injury in rats.
Molecular medicine reports
Li Z,Liu F,Zhang L,Cao Y,Shao Y,Wang X,Jiang X,Chen Z
Thu Feb 01 00:00:00 EST 2018
Neuroserpin restores autophagy and promotes functional recovery after acute spinal cord injury in rats.
Molecular medicine reports
Li Z,Liu F,Zhang L,Cao Y,Shao Y,Wang X,Jiang X,Chen Z
Thu Feb 01 00:00:00 EST 2018
In Vitro Neural Differentiation of Bone Marrow Mesenchymal Stem Cells Carrying the FTH1 Reporter Gene and Detection with MRI.
BioMed research international
Mu T,Qin Y,Liu B,He X,Liao Y,Sun J,Qiu J,Li X,Zhong Y,Cai J
Mon Jan 14 00:00:00 EST 2019
In Vitro Neural Differentiation of Bone Marrow Mesenchymal Stem Cells Carrying the FTH1 Reporter Gene and Detection with MRI.
BioMed research international
Mu T,Qin Y,Liu B,He X,Liao Y,Sun J,Qiu J,Li X,Zhong Y,Cai J
Mon Jan 14 00:00:00 EST 2019
Pitavastatin treatment induces neuroprotection through the BDNF-TrkB signalling pathway in cultured cerebral neurons after oxygen-glucose deprivation.
Neurological research
Cui X,Fu Z,Wang M,Nan X,Zhang B
Tue May 01 00:00:00 EDT 2018
Pitavastatin treatment induces neuroprotection through the BDNF-TrkB signalling pathway in cultured cerebral neurons after oxygen-glucose deprivation.
Neurological research
Cui X,Fu Z,Wang M,Nan X,Zhang B
Tue May 01 00:00:00 EDT 2018
REST overexpression in mice causes deficits in spontaneous locomotion.
Scientific reports
Lu L,Marisetty A,Liu B,Kamal MM,Gumin J,Veo B,Cai Y,Kassem DH,Weng C,Maynard ME,Hood KN,Fuller GN,Pan ZZ,Cykowski MD,Dash PK,Majumder S
Tue Aug 14 00:00:00 EDT 2018
REST overexpression in mice causes deficits in spontaneous locomotion.
Scientific reports
Lu L,Marisetty A,Liu B,Kamal MM,Gumin J,Veo B,Cai Y,Kassem DH,Weng C,Maynard ME,Hood KN,Fuller GN,Pan ZZ,Cykowski MD,Dash PK,Majumder S
Tue Aug 14 00:00:00 EDT 2018
Downregulated Nuclear Factor E2-Related Factor 2 (Nrf2) Aggravates Cognitive Impairments via Neuroinflammation and Synaptic Plasticity in the Senescence-Accelerated Mouse Prone 8 (SAMP8) Mouse: A Model of Accelerated Senescence.
Medical science monitor : international medical journal of experimental and clinical research
Ren HL,Lv CN,Xing Y,Geng Y,Zhang F,Bu W,Wang MW
Fri Feb 23 00:00:00 EST 2018
Downregulated Nuclear Factor E2-Related Factor 2 (Nrf2) Aggravates Cognitive Impairments via Neuroinflammation and Synaptic Plasticity in the Senescence-Accelerated Mouse Prone 8 (SAMP8) Mouse: A Model of Accelerated Senescence.
Medical science monitor : international medical journal of experimental and clinical research
Ren HL,Lv CN,Xing Y,Geng Y,Zhang F,Bu W,Wang MW
Fri Feb 23 00:00:00 EST 2018
Carbon monoxide-releasing molecule-3 protects against ischemic stroke by suppressing neuroinflammation and alleviating blood-brain barrier disruption.
Journal of neuroinflammation
Wang J,Zhang D,Fu X,Yu L,Lu Z,Gao Y,Liu X,Man J,Li S,Li N,Chen X,Hong M,Yang Q,Wang J
Thu Jun 21 00:00:00 EDT 2018
Carbon monoxide-releasing molecule-3 protects against ischemic stroke by suppressing neuroinflammation and alleviating blood-brain barrier disruption.
Journal of neuroinflammation
Wang J,Zhang D,Fu X,Yu L,Lu Z,Gao Y,Liu X,Man J,Li S,Li N,Chen X,Hong M,Yang Q,Wang J
Thu Jun 21 00:00:00 EDT 2018
Mutant huntingtin protein expression and blood-spinal cord barrier dysfunction in huntington disease.
Annals of neurology
Sciacca G,Cicchetti F
Fri Dec 01 00:00:00 EST 2017
Mutant huntingtin protein expression and blood-spinal cord barrier dysfunction in huntington disease.
Annals of neurology
Sciacca G,Cicchetti F
Fri Dec 01 00:00:00 EST 2017
RNA stores tau reversibly in complex coacervates.
PLoS biology
Zhang X,Lin Y,Eschmann NA,Zhou H,Rauch JN,Hernandez I,Guzman E,Kosik KS,Han S
Sat Jul 01 00:00:00 EDT 2017
RNA stores tau reversibly in complex coacervates.
PLoS biology
Zhang X,Lin Y,Eschmann NA,Zhou H,Rauch JN,Hernandez I,Guzman E,Kosik KS,Han S
Sat Jul 01 00:00:00 EDT 2017
The effects of fluoride on neuronal function occurs via cytoskeleton damage and decreased signal transmission.
Chemosphere
Chen L,Ning H,Yin Z,Song X,Feng Y,Qin H,Li Y,Wang J,Ge Y,Wang W
Sun Oct 01 00:00:00 EDT 2017
The effects of fluoride on neuronal function occurs via cytoskeleton damage and decreased signal transmission.
Chemosphere
Chen L,Ning H,Yin Z,Song X,Feng Y,Qin H,Li Y,Wang J,Ge Y,Wang W
Sun Oct 01 00:00:00 EDT 2017
Regulation of the blood-testis barrier by a local axis in the testis: role of laminin α2 in the basement membrane.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Gao Y,Mruk D,Chen H,Lui WY,Lee WM,Cheng CY
Wed Feb 01 00:00:00 EST 2017
Regulation of the blood-testis barrier by a local axis in the testis: role of laminin α2 in the basement membrane.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Gao Y,Mruk D,Chen H,Lui WY,Lee WM,Cheng CY
Wed Feb 01 00:00:00 EST 2017
miR-200c regulates endothelin-1 induced PASMCs abnormal proliferation and apoptosis.
IUBMB life
Yuan C,Xu M,Rong R,Mei Y,Cai W,Li L,Xue Y,Zhu B,Sun K,Han L
Wed Nov 01 00:00:00 EDT 2017
miR-200c regulates endothelin-1 induced PASMCs abnormal proliferation and apoptosis.
IUBMB life
Yuan C,Xu M,Rong R,Mei Y,Cai W,Li L,Xue Y,Zhu B,Sun K,Han L
Wed Nov 01 00:00:00 EDT 2017
Transplantation of VEGF-mediated bone marrow mesenchymal stem cells promotes functional improvement in a rat acute cerebral infarction model.
Brain research
Zong X,Wu S,Li F,Lv L,Han D,Zhao N,Yan X,Hu S,Xu T
Fri Dec 01 00:00:00 EST 2017
Transplantation of VEGF-mediated bone marrow mesenchymal stem cells promotes functional improvement in a rat acute cerebral infarction model.
Brain research
Zong X,Wu S,Li F,Lv L,Han D,Zhao N,Yan X,Hu S,Xu T
Fri Dec 01 00:00:00 EST 2017
Transplantation of VEGF-mediated bone marrow mesenchymal stem cells promotes functional improvement in a rat acute cerebral infarction model.
Brain research
Zong X,Wu S,Li F,Lv L,Han D,Zhao N,Yan X,Hu S,Xu T
Fri Dec 01 00:00:00 EST 2017
Transplantation of VEGF-mediated bone marrow mesenchymal stem cells promotes functional improvement in a rat acute cerebral infarction model.
Brain research
Zong X,Wu S,Li F,Lv L,Han D,Zhao N,Yan X,Hu S,Xu T
Fri Dec 01 00:00:00 EST 2017
Involvement of intracellular transport in TREK-1c current run-up in 293T cells.
Channels (Austin, Tex.)
Andharia N,Joseph A,Hayashi M,Okada M,Matsuda H
Thu May 04 00:00:00 EDT 2017
Involvement of intracellular transport in TREK-1c current run-up in 293T cells.
Channels (Austin, Tex.)
Andharia N,Joseph A,Hayashi M,Okada M,Matsuda H
Thu May 04 00:00:00 EDT 2017
Anterograde monosynaptic transneuronal tracers derived from herpes simplex virus 1 strain H129.
Molecular neurodegeneration
Zeng WB,Jiang HF,Gang YD,Song YG,Shen ZZ,Yang H,Dong X,Tian YL,Ni RJ,Liu Y,Tang N,Li X,Jiang X,Gao D,Androulakis M,He XB,Xia HM,Ming YZ,Lu Y,Zhou JN,Zhang C,Xia XS,Shu Y,Zeng SQ,Xu F,Zhao F,Luo MH
Fri May 12 00:00:00 EDT 2017
Anterograde monosynaptic transneuronal tracers derived from herpes simplex virus 1 strain H129.
Molecular neurodegeneration
Zeng WB,Jiang HF,Gang YD,Song YG,Shen ZZ,Yang H,Dong X,Tian YL,Ni RJ,Liu Y,Tang N,Li X,Jiang X,Gao D,Androulakis M,He XB,Xia HM,Ming YZ,Lu Y,Zhou JN,Zhang C,Xia XS,Shu Y,Zeng SQ,Xu F,Zhao F,Luo MH
Fri May 12 00:00:00 EDT 2017
rhEPO Enhances Cellular Anti-oxidant Capacity to Protect Long-Term Cultured Aging Primary Nerve Cells.
Journal of molecular neuroscience : MN
Wang H,Fan J,Chen M,Yao Q,Gao Z,Zhang G,Wu H,Yu X
Tue Aug 01 00:00:00 EDT 2017
Recent Zika Virus Isolates Induce Premature Differentiation of Neural Progenitors in Human Brain Organoids.
Cell stem cell
Gabriel E,Ramani A,Karow U,Gottardo M,Natarajan K,Gooi LM,Goranci-Buzhala G,Krut O,Peters F,Nikolic M,Kuivanen S,Korhonen E,Smura T,Vapalahti O,Papantonis A,Schmidt-Chanasit J,Riparbelli M,Callaini G,Krönke M,Utermöhlen O,Gopalakrishnan J
Thu Mar 02 00:00:00 EST 2017
Recent Zika Virus Isolates Induce Premature Differentiation of Neural Progenitors in Human Brain Organoids.
Cell stem cell
Gabriel E,Ramani A,Karow U,Gottardo M,Natarajan K,Gooi LM,Goranci-Buzhala G,Krut O,Peters F,Nikolic M,Kuivanen S,Korhonen E,Smura T,Vapalahti O,Papantonis A,Schmidt-Chanasit J,Riparbelli M,Callaini G,Krönke M,Utermöhlen O,Gopalakrishnan J
Thu Mar 02 00:00:00 EST 2017
An Important Function of Petrosiol E in Inducing the Differentiation of Neuronal Progenitors and in Protecting Them against Oxidative Stress.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Liu J,Wang L,Du Y,Liu S
Sun Oct 01 00:00:00 EDT 2017
An Important Function of Petrosiol E in Inducing the Differentiation of Neuronal Progenitors and in Protecting Them against Oxidative Stress.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Liu J,Wang L,Du Y,Liu S
Sun Oct 01 00:00:00 EDT 2017
Improved Neural Regeneration with Olfactory Ensheathing Cell Inoculated PLGA Scaffolds in Spinal Cord Injury Adult Rats.
Neuro-Signals
Wang C,Sun C,Hu Z,Huo X,Yang Y,Liu X,Botchway BOA,Davies H,Fang M
Tue Mar 05 00:00:00 EST 2019
Improved Neural Regeneration with Olfactory Ensheathing Cell Inoculated PLGA Scaffolds in Spinal Cord Injury Adult Rats.
Neuro-Signals
Wang C,Sun C,Hu Z,Huo X,Yang Y,Liu X,Botchway BOA,Davies H,Fang M
Tue Mar 05 00:00:00 EST 2019
Human-to-mouse prion-like propagation of mutant huntingtin protein.
Acta neuropathologica
Jeon I,Cicchetti F,Cisbani G,Lee S,Li E,Bae J,Lee N,Li L,Im W,Kim M,Kim HS,Oh SH,Kim TA,Ko JJ,Aubé B,Oueslati A,Kim YJ,Song J
Sat Oct 01 00:00:00 EDT 2016
Human-to-mouse prion-like propagation of mutant huntingtin protein.
Acta neuropathologica
Jeon I,Cicchetti F,Cisbani G,Lee S,Li E,Bae J,Lee N,Li L,Im W,Kim M,Kim HS,Oh SH,Kim TA,Ko JJ,Aubé B,Oueslati A,Kim YJ,Song J
Sat Oct 01 00:00:00 EDT 2016
Gadd45b prevents autophagy and apoptosis against rat cerebral neuron oxygen-glucose deprivation/reperfusion injury.
Apoptosis : an international journal on programmed cell death
He G,Xu W,Tong L,Li S,Su S,Tan X,Li C
Fri Apr 01 00:00:00 EDT 2016
Gadd45b prevents autophagy and apoptosis against rat cerebral neuron oxygen-glucose deprivation/reperfusion injury.
Apoptosis : an international journal on programmed cell death
He G,Xu W,Tong L,Li S,Su S,Tan X,Li C
Fri Apr 01 00:00:00 EDT 2016
Claulansine F promoted the neuronal differentiation of neural stem and progenitor cells through Akt/GSK-3β/β-catenin pathway.
European journal of pharmacology
Huang JY,Ma YZ,Yuan YH,Zuo W,Chu SF,Liu H,Du GH,Zhang DM,Chen NH
Mon Sep 05 00:00:00 EDT 2016
Claulansine F promoted the neuronal differentiation of neural stem and progenitor cells through Akt/GSK-3β/β-catenin pathway.
European journal of pharmacology
Huang JY,Ma YZ,Yuan YH,Zuo W,Chu SF,Liu H,Du GH,Zhang DM,Chen NH
Mon Sep 05 00:00:00 EDT 2016
Claulansine F promoted the neuronal differentiation of neural stem and progenitor cells through Akt/GSK-3β/β-catenin pathway.
European journal of pharmacology
Huang JY,Ma YZ,Yuan YH,Zuo W,Chu SF,Liu H,Du GH,Zhang DM,Chen NH
Mon Sep 05 00:00:00 EDT 2016
Claulansine F promoted the neuronal differentiation of neural stem and progenitor cells through Akt/GSK-3β/β-catenin pathway.
European journal of pharmacology
Huang JY,Ma YZ,Yuan YH,Zuo W,Chu SF,Liu H,Du GH,Zhang DM,Chen NH
Mon Sep 05 00:00:00 EDT 2016
Endoplasmic reticulum stress mediates JNK-dependent IRS-1 serine phosphorylation and results in Tau hyperphosphorylation in amyloid β oligomer-treated PC12 cells and primary neurons.
Gene
Zhang X,Tang S,Zhang Q,Shao W,Han X,Wang Y,Du Y
Wed Aug 10 00:00:00 EDT 2016
Endoplasmic reticulum stress mediates JNK-dependent IRS-1 serine phosphorylation and results in Tau hyperphosphorylation in amyloid β oligomer-treated PC12 cells and primary neurons.
Gene
Zhang X,Tang S,Zhang Q,Shao W,Han X,Wang Y,Du Y
Wed Aug 10 00:00:00 EDT 2016
Differential epigenetic effects of chlorpyrifos and arsenic in proliferating and differentiating human neural progenitor cells.
Reproductive toxicology (Elmsford, N.Y.)
Kim HY,Wegner SH,Van Ness KP,Park JJ,Pacheco SE,Workman T,Hong S,Griffith W,Faustman EM
Sat Oct 01 00:00:00 EDT 2016
Differential epigenetic effects of chlorpyrifos and arsenic in proliferating and differentiating human neural progenitor cells.
Reproductive toxicology (Elmsford, N.Y.)
Kim HY,Wegner SH,Van Ness KP,Park JJ,Pacheco SE,Workman T,Hong S,Griffith W,Faustman EM
Sat Oct 01 00:00:00 EDT 2016
Evaluation of histopathological changes in the microstructure at the center and periphery of glioma tumors using diffusional kurtosis imaging.
Clinical neurology and neurosurgery
Li F,Shi W,Wang D,Xu Y,Li H,He J,Zeng Q
Thu Dec 01 00:00:00 EST 2016
Viral Infection of the Central Nervous System and Neuroinflammation Precede Blood-Brain Barrier Disruption during Japanese Encephalitis Virus Infection.
Journal of virology
Li F,Wang Y,Yu L,Cao S,Wang K,Yuan J,Wang C,Wang K,Cui M,Fu ZF
Fri May 01 00:00:00 EDT 2015
Viral Infection of the Central Nervous System and Neuroinflammation Precede Blood-Brain Barrier Disruption during Japanese Encephalitis Virus Infection.
Journal of virology
Li F,Wang Y,Yu L,Cao S,Wang K,Yuan J,Wang C,Wang K,Cui M,Fu ZF
Fri May 01 00:00:00 EDT 2015
Headless Myo10 is a regulator of microtubule stability during neuronal development.
Journal of neurochemistry
Yu H,Sun D,Wang N,Wang M,Lan Y,Fan W,Zhao Y,Guo W,Zhu X
Thu Oct 01 00:00:00 EDT 2015
Headless Myo10 is a regulator of microtubule stability during neuronal development.
Journal of neurochemistry
Yu H,Sun D,Wang N,Wang M,Lan Y,Fan W,Zhao Y,Guo W,Zhu X
Thu Oct 01 00:00:00 EDT 2015
Increased stathmin expression strengthens fear conditioning in epileptic rats.
Biomedical reports
Zhang L,Feng D,Tao H,DE X,Chang Q,Hu Q
Thu Jan 01 00:00:00 EST 2015
Increased stathmin expression strengthens fear conditioning in epileptic rats.
Biomedical reports
Zhang L,Feng D,Tao H,DE X,Chang Q,Hu Q
Thu Jan 01 00:00:00 EST 2015
Huwe1 interacts with Gadd45b under oxygen-glucose deprivation and reperfusion injury in primary Rat cortical neuronal cells.
Molecular brain
He GQ,Xu WM,Li JF,Li SS,Liu B,Tan XD,Li CQ
Wed Dec 23 00:00:00 EST 2015
Huwe1 interacts with Gadd45b under oxygen-glucose deprivation and reperfusion injury in primary Rat cortical neuronal cells.
Molecular brain
He GQ,Xu WM,Li JF,Li SS,Liu B,Tan XD,Li CQ
Wed Dec 23 00:00:00 EST 2015
Mutant huntingtin is present in neuronal grafts in Huntington disease patients.
Annals of neurology
Cicchetti F,Lacroix S,Cisbani G,Vallières N,Saint-Pierre M,St-Amour I,Tolouei R,Skepper JN,Hauser RA,Mantovani D,Barker RA,Freeman TB
Tue Jul 01 00:00:00 EDT 2014
Mutant huntingtin is present in neuronal grafts in Huntington disease patients.
Annals of neurology
Cicchetti F,Lacroix S,Cisbani G,Vallières N,Saint-Pierre M,St-Amour I,Tolouei R,Skepper JN,Hauser RA,Mantovani D,Barker RA,Freeman TB
Tue Jul 01 00:00:00 EDT 2014
Neuropeptide Y protects cerebral cortical neurons by regulating microglial immune function.
Neural regeneration research
Li Q,Dong C,Li W,Bu W,Wu J,Zhao W
Thu May 01 00:00:00 EDT 2014
Neuropeptide Y protects cerebral cortical neurons by regulating microglial immune function.
Neural regeneration research
Li Q,Dong C,Li W,Bu W,Wu J,Zhao W
Thu May 01 00:00:00 EDT 2014
Bone marrow mesenchymal stem cells repair spinal cord ischemia/reperfusion injury by promoting axonal growth and anti-autophagy.
Neural regeneration research
Yin F,Meng C,Lu R,Li L,Zhang Y,Chen H,Qin Y,Guo L
Mon Sep 15 00:00:00 EDT 2014
Bone marrow mesenchymal stem cells repair spinal cord ischemia/reperfusion injury by promoting axonal growth and anti-autophagy.
Neural regeneration research
Yin F,Meng C,Lu R,Li L,Zhang Y,Chen H,Qin Y,Guo L
Mon Sep 15 00:00:00 EDT 2014
Transplantation of mesenchymal stem cells exerts anti-apoptotic effects in adult rats after spinal cord ischemia-reperfusion injury.
Brain research
Yin F,Guo L,Meng CY,Liu YJ,Lu RF,Li P,Zhou YB
Fri May 02 00:00:00 EDT 2014
Transplantation of mesenchymal stem cells exerts anti-apoptotic effects in adult rats after spinal cord ischemia-reperfusion injury.
Brain research
Yin F,Guo L,Meng CY,Liu YJ,Lu RF,Li P,Zhou YB
Fri May 02 00:00:00 EDT 2014
AxonQuant: A Microfluidic Chamber Culture-Coupled Algorithm That Allows High-Throughput Quantification of Axonal Damage.
Neuro-Signals
Li Y,Yang M,Huang Z,Chen X,Maloney MT,Zhu L,Liu J,Yang Y,Du S,Jiang X,Wu JY
Wed Oct 14 00:00:00 EDT 2015
AxonQuant: A Microfluidic Chamber Culture-Coupled Algorithm That Allows High-Throughput Quantification of Axonal Damage.
Neuro-Signals
Li Y,Yang M,Huang Z,Chen X,Maloney MT,Zhu L,Liu J,Yang Y,Du S,Jiang X,Wu JY
Wed Oct 14 00:00:00 EDT 2015
The effects of harvesting media on biological characteristics and repair potential of neural stem cells after traumatic brain injury.
PloS one
Liu S,Li Z,Fu J,Sun L,Xu F,Harada T,Lou Y,Chu M,Sun Q,Xu K,Zhang R,Jin L,Xiao H,Wu S
Mon Jun 22 00:00:00 EDT 2015
The effects of harvesting media on biological characteristics and repair potential of neural stem cells after traumatic brain injury.
PloS one
Liu S,Li Z,Fu J,Sun L,Xu F,Harada T,Lou Y,Chu M,Sun Q,Xu K,Zhang R,Jin L,Xiao H,Wu S
Mon Jun 22 00:00:00 EDT 2015
Effects of targeted suppression of glutaryl-CoA dehydrogenase by lentivirus-mediated shRNA and excessive intake of lysine on apoptosis in rat striatal neurons.
PloS one
Gao J,Zhang C,Fu X,Yi Q,Tian F,Ning Q,Luo X
Wed Dec 04 00:00:00 EST 2013
Effects of targeted suppression of glutaryl-CoA dehydrogenase by lentivirus-mediated shRNA and excessive intake of lysine on apoptosis in rat striatal neurons.
PloS one
Gao J,Zhang C,Fu X,Yi Q,Tian F,Ning Q,Luo X
Wed Dec 04 00:00:00 EST 2013
Effects of targeted suppression of glutaryl-CoA dehydrogenase by lentivirus-mediated shRNA and excessive intake of lysine on apoptosis in rat striatal neurons.
PloS one
Gao J,Zhang C,Fu X,Yi Q,Tian F,Ning Q,Luo X
Wed Dec 04 00:00:00 EST 2013
Effects of targeted suppression of glutaryl-CoA dehydrogenase by lentivirus-mediated shRNA and excessive intake of lysine on apoptosis in rat striatal neurons.
PloS one
Gao J,Zhang C,Fu X,Yi Q,Tian F,Ning Q,Luo X
Wed Dec 04 00:00:00 EST 2013