Product References
Human IRF1 governs macrophagic IFN-γ immunity to mycobacteria.
Cell
Rosain J,Neehus AL,Manry J,Yang R,Le Pen J,Daher W,Liu Z,Chan YH,Tahuil N,Türel Ö,Bourgey M,Ogishi M,Doisne JM,Izquierdo HM,Shirasaki T,Le Voyer T,Guérin A,Bastard P,Moncada-Vélez M,Han JE,Khan T,Rapaport F,Hong SH,Cheung A,Haake K,Mindt BC,Pérez L,Philippot Q,Lee D,Zhang P,Rinchai D,Al Ali F,Ahmad Ata MM,Rahman M,Peel JN,Heissel S,Molina H,Kendir-Demirkol Y,Bailey R,Zhao S,Bohlen J,Mancini M,Seeleuthner Y,Roelens M,Lorenzo L,Soudée C,Paz MEJ,González ML,Jeljeli M,Soulier J,Romana S,L'Honneur AS,Materna M,Martínez-Barricarte R,Pochon M,Oleaga-Quintas C,Michev A,Migaud M,Lévy R,Alyanakian MA,Rozenberg F,Croft CA,Vogt G,Emile JF,Kremer L,Ma CS,Fritz JH,Lemon SM,Spaan AN,Manel N,Abel L,MacDonald MR,Boisson-Dupuis S,Marr N,Tangye SG,Di Santo JP,Zhang Q,Zhang SY,Rice CM,Béziat V,Lachmann N,Langlais D,Casanova JL,Gros P,Bustamante J
Thu Feb 02 00:00:00 EST 2023
The antiviral activity of tripartite motif protein 38 in hepatitis B virus replication and gene expression and its association with treatment responses during PEG-IFN-α antiviral therapy.
Virology
Luo H,Hu X,Li Y,Lei D,Tan G,Zeng Y,Qin B
Wed Feb 01 00:00:00 EST 2023
Transcriptomic and antiviral analyses of PoIFN-Delta5 against porcine enteric viruses in porcine intestinal epithelial cells.
Veterinary microbiology
Zhu X,Fan B,Song S,Gao J,Zhou J,Zhao Y,Guo W,Zhang X,Qian J,Zhu M,Li J,Li Y,Guo R,Fan H,Dong H,Zhang X,Li B
Mon May 01 00:00:00 EDT 2023
Intranasal or airborne transmission-mediated delivery of an attenuated SARS-CoV-2 protects Syrian hamsters against new variants.
Nature communications
Stauft CB,Selvaraj P,D'Agnillo F,Meseda CA,Liu S,Pedro CL,Sangare K,Lien CZ,Weir JP,Starost MF,Wang TT
Fri Jun 09 00:00:00 EDT 2023
Acquired chemoresistance can lead to increased resistance of pancreatic cancer cells to oncolytic vesicular stomatitis virus.
Molecular therapy oncolytics
Goad DW,Bressy C,Holbrook MC,Grdzelishvili VZ
Thu Mar 17 00:00:00 EDT 2022
Anti-inflammatory Properties of the Alpha-Melanocyte-Stimulating Hormone in Models of Granulomatous Inflammation.
Lung
Shahraki AH,Tian R,Zhang C,Fregien NL,Bejarano P,Mirsaeidi M
Mon Aug 01 00:00:00 EDT 2022
Active and Passive Immunization of Syrian Hamsters with An Attenuated SARS-CoV-2 Protects against New Variants of Concern.
Research square
Wang T,Stauft C,Selvaraj P,D'agnillo F,Meseda C,Sangare K,Pedro C,Liu S,Lien C,Weir J,Starost M
Fri Nov 11 00:00:00 EST 2022
Marek's Disease Virus and Reticuloendotheliosis Virus Coinfection Enhances Viral Replication and Alters Cellular Protein Profiles.
Frontiers in veterinary science
Du X,Zhou D,Zhou J,Xue J,Cheng Z
Sat Apr 09 00:00:00 EDT 2022
Proteomic Investigation Reveals Eukaryotic Translation Initiation Factor 5A Involvement in Porcine Reproductive and Respiratory Syndrome Virus Infection in vitro.
Frontiers in veterinary science
Li H,Wan B,Jiang D,Ji P,Zhao M,Li X,Li R,Qiao S
Tue May 03 00:00:00 EDT 2022
Temporally integrated transcriptome analysis reveals ASFV pathology and host response dynamics.
Frontiers in immunology
Lv L,Zhang T,Jia H,Zhang Y,Ahsan A,Zhao X,Chen T,Shen Z,Shen N
Fri Dec 23 00:00:00 EST 2022
LncRNA NEAT1 Potentiates SREBP2 Activity to Promote Inflammatory Macrophage Activation and Limit Hantaan Virus Propagation.
Frontiers in microbiology
Yang Y,Li M,Ma Y,Ye W,Si Y,Zheng X,Liu H,Cheng L,Zhang L,Zhang H,Zhang X,Lei Y,Shen L,Zhang F,Ma H
Sat Jul 16 00:00:00 EDT 2022
The Type I Interferon Pathway Is Upregulated in the Cutaneous Lesions and Blood of Multibacillary Leprosy Patients With Erythema Nodosum Leprosum.
Frontiers in medicine
Rosa TLSA,Mendes MA,Linhares NRC,Rodrigues TF,Dias AA,Leal-Calvo T,Gandini M,Ferreira H,Costa FDMR,Sales AM,Amadeu TP,Schmitz V,Pinheiro RO,Rodrigues LS,Moraes MO,Pessolani MCV
Sat Jul 16 00:00:00 EDT 2022
Single-cell profiling reveals distinct adaptive immune hallmarks in MDA5+ dermatomyositis with therapeutic implications.
Nature communications
Ye Y,Chen Z,Jiang S,Jia F,Li T,Lu X,Xue J,Lian X,Ma J,Hao P,Lu L,Ye S,Shen N,Bao C,Fu Q,Zhang X
Sat Oct 29 00:00:00 EDT 2022
Regulation of type I interferon signature by VGLL3 in the fibroblast-like synoviocytes of rheumatoid arthritis patients via targeting the Hippo pathway.
Arthritis research & therapy
Du Y,Cui R,Tian N,Chen M,Zhang XL,Dai SM
Mon Aug 08 00:00:00 EDT 2022
Regulation of type I interferon signature by VGLL3 in the fibroblast-like synoviocytes of rheumatoid arthritis patients via targeting the Hippo pathway.
Arthritis research & therapy
Du Y,Cui R,Tian N,Chen M,Zhang XL,Dai SM
Mon Aug 08 00:00:00 EDT 2022
PIAS1-mediated SUMOylation of influenza A virus PB2 restricts viral replication and virulence.
PLoS pathogens
Wang G,Zhao Y,Zhou Y,Jiang L,Liang L,Kong F,Yan Y,Wang X,Wang Y,Wen X,Zeng X,Tian G,Deng G,Shi J,Liu L,Chen H,Li C
Fri Apr 01 00:00:00 EDT 2022
Interferon-induced degradation of the persistent hepatitis B virus cccDNA form depends on ISG20.
EMBO reports
Stadler D,Kächele M,Jones AN,Hess J,Urban C,Schneider J,Xia Y,Oswald A,Nebioglu F,Bester R,Lasitschka F,Ringelhan M,Ko C,Chou WM,Geerlof A,van de Klundert MA,Wettengel JM,Schirmacher P,Heikenwälder M,Schreiner S,Bartenschlager R,Pichlmair A,Sattler M,Unger K,Protzer U
Fri Jun 04 00:00:00 EDT 2021
RhoC Modulates Cell Junctions and Type I Interferon Response in Aggressive Breast Cancers.
Frontiers in oncology
Abraham HG,Ulintz PJ,Goo L,Yates JA,Little AC,Bao L,Wu Z,Merajver SD
Tue Nov 07 00:00:00 EST 2023
Inducible ATP1B1 Upregulates Antiviral Innate Immune Responses by the Ubiquitination of TRAF3 and TRAF6.
Journal of immunology (Baltimore, Md. : 1950)
Cao W,Guo Y,Cheng Z,Xu G,Zuo Q,Nie L,Huang Y,Liu S,Zhu Y
Tue Jun 01 00:00:00 EDT 2021
The ApoA-I mimetic peptide 4F attenuates in vitro replication of SARS-CoV-2, associated apoptosis, oxidative stress and inflammation in epithelial cells.
Virulence
Kelesidis T,Madhav S,Petcherski A,Cristelle H,O'Connor E,Hultgren NW,Ritou E,Williams DS,Shirihai OS,Reddy ST
Wed Dec 01 00:00:00 EST 2021
Human parainfluenza virus type 1 regulates cholesterol biosynthesis and establishes quiescent infection in human airway cells.
PLoS pathogens
Kurebayashi Y,Bajimaya S,Watanabe M,Lim N,Lutz M,Dunagan M,Takimoto T
Wed Sep 01 00:00:00 EDT 2021
Human parainfluenza virus type 1 regulates cholesterol biosynthesis and establishes quiescent infection in human airway cells.
PLoS pathogens
Kurebayashi Y,Bajimaya S,Watanabe M,Lim N,Lutz M,Dunagan M,Takimoto T
Wed Sep 01 00:00:00 EDT 2021
Virus-inducible IGFALS facilitates innate immune responses by mediating IRAK1 and TRAF6 activation.
Cellular & molecular immunology
Xu G,Deng F,Zuo Q,Liu L,Dou K,Cheng Z,Cao W,Luo C,Yu C,Liu S,Zhu Y
Tue Jun 01 00:00:00 EDT 2021
Large-scale and high-resolution mass spectrometry-based proteomics profiling defines molecular subtypes of esophageal cancer for therapeutic targeting.
Nature communications
Liu W,Xie L,He YH,Wu ZY,Liu LX,Bai XF,Deng DX,Xu XE,Liao LD,Lin W,Heng JH,Xu X,Peng L,Huang QF,Li CY,Zhang ZD,Wang W,Zhang GR,Gao X,Wang SH,Li CQ,Xu LY,Liu W,Li EM
Mon Aug 16 00:00:00 EDT 2021
Role of IL-6-IL-27 Complex in Host Antiviral Immune Response.
Journal of immunology (Baltimore, Md. : 1950)
Zuo Q,Cheng Z,Zhang G,Xia Y,Xu G,Cao W,Yang X,Fu Y,He R,Fang P,Guo Y,Nie L,Huang Y,Liu L,Zhan J,Liu S,Zhu Y
Thu Jul 15 00:00:00 EDT 2021
The Protein Kinase Receptor Modulates the Innate Immune Response against Tacaribe Virus.
Viruses
Moreno H,Kunz S
Wed Jul 07 00:00:00 EDT 2021
The G Protein-Coupled Receptor FFAR2 Promotes Internalization during Influenza A Virus Entry.
Journal of virology
Wang G,Jiang L,Wang J,Zhang J,Kong F,Li Q,Yan Y,Huang S,Zhao Y,Liang L,Li J,Sun N,Hu Y,Shi W,Deng G,Chen P,Liu L,Zeng X,Tian G,Bu Z,Chen H,Li C
Mon Jan 06 00:00:00 EST 2020
SFTSV Infection Induces BAK/BAX-Dependent Mitochondrial DNA Release to Trigger NLRP3 Inflammasome Activation.
Cell reports
Li S,Li H,Zhang YL,Xin QL,Guan ZQ,Chen X,Zhang XA,Li XK,Xiao GF,Lozach PY,Cui J,Liu W,Zhang LK,Peng K
Tue Mar 31 00:00:00 EDT 2020
A RIG-I-like receptor directs antiviral responses to a bunyavirus and is antagonized by virus-induced blockade of TRIM25-mediated ubiquitination.
The Journal of biological chemistry
Min YQ,Ning YJ,Wang H,Deng F
Fri Jul 10 00:00:00 EDT 2020
Cross-talk between SUMOylation and ISGylation in response to interferon.
Cytokine
El-Asmi F,McManus FP,Brantis-de-Carvalho CE,Valle-Casuso JC,Thibault P,Chelbi-Alix MK
Fri May 01 00:00:00 EDT 2020
Exosomal miR-145-5p derived from orthohantavirus-infected endothelial cells inhibits HTNV infection.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Wang X,Chen QZ,Zan YX,Wang MR,Yan J,Guo WW,Li KJ,Liu YY,Luo F,Feng Y,Hou W,Xiong HR
Thu Oct 01 00:00:00 EDT 2020
Direct Antiviral Activity of IFN-Stimulated Genes Is Responsible for Resistance to Paramyxoviruses in ISG15-Deficient Cells.
Journal of immunology (Baltimore, Md. : 1950)
Holthaus D,Vasou A,Bamford CGG,Andrejeva J,Paulus C,Randall RE,McLauchlan J,Hughes DJ
Wed Jul 01 00:00:00 EDT 2020
Induction of interleukin 6 impairs the anti-HBV efficiency of IFN-α in human hepatocytes through upregulation of SOCS3.
Journal of medical virology
Yang K,Guan S,Zhang H,Chen Z
Wed May 01 00:00:00 EDT 2019
Induction of interleukin 6 impairs the anti-HBV efficiency of IFN-α in human hepatocytes through upregulation of SOCS3.
Journal of medical virology
Yang K,Guan S,Zhang H,Chen Z
Wed May 01 00:00:00 EDT 2019
Interferon-alpha promotes immunosuppression through IFNAR1/STAT1 signalling in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Yang W,Zhang L,Liu S,Zhao M,Zhou G,Wang L,Jin S,Zhang Z,Hu J
Fri Feb 01 00:00:00 EST 2019
Interferon-alpha promotes immunosuppression through IFNAR1/STAT1 signalling in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Yang W,Zhang L,Liu S,Zhao M,Zhou G,Wang L,Jin S,Zhang Z,Hu J
Fri Feb 01 00:00:00 EST 2019
Characterization, functional and signaling elucidation of pigeon (Columba livia) interferon-α: Knockdown p53 negatively modulates antiviral response.
Developmental and comparative immunology
Li S,Wang Y,Zhao H,Shao Y,Liu J,Xing M
Tue Jan 01 00:00:00 EST 2019
Inducible LGALS3BP/90K activates antiviral innate immune responses by targeting TRAF6 and TRAF3 complex.
PLoS pathogens
Xu G,Xia Z,Deng F,Liu L,Wang Q,Yu Y,Wang F,Zhu C,Liu W,Cheng Z,Zhu Y,Zhou L,Zhang Y,Lu M,Liu S
Thu Aug 01 00:00:00 EDT 2019
Docosahexaenoic Acid Consumption Impedes Early Interferon- and Chemokine-Related Gene Expression While Suppressing Silica-Triggered Flaring of Murine Lupus.
Frontiers in immunology
Benninghoff AD,Bates MA,Chauhan PS,Wierenga KA,Gilley KN,Holian A,Harkema JR,Pestka JJ
Wed Nov 04 00:00:00 EST 2020
Molecular cloning and transcriptional regulation of Indian peafowl (Pavo cristatus) IFN-α gene.
Cell stress & chaperones
Wang Y,Zhao H,Liu J,Shao Y,Xing M
Fri Mar 01 00:00:00 EST 2019
Comparative Analysis of the Antiviral Effects Mediated by Type I and III Interferons in Hepatitis B Virus-Infected Hepatocytes.
The Journal of infectious diseases
Bockmann JH,Stadler D,Xia Y,Ko C,Wettengel JM,Schulze Zur Wiesch J,Dandri M,Protzer U
Fri Jul 19 00:00:00 EDT 2019
Long Noncoding RNA Lnc-MxA Inhibits Beta Interferon Transcription by Forming RNA-DNA Triplexes at Its Promoter.
Journal of virology
Li X,Guo G,Lu M,Chai W,Li Y,Tong X,Li J,Jia X,Liu W,Qi D,Ye X
Fri Nov 01 00:00:00 EDT 2019
The histone chaperone HIRA promotes the induction of host innate immune defences in response to HSV-1 infection.
PLoS pathogens
McFarlane S,Orr A,Roberts APE,Conn KL,Iliev V,Loney C,da Silva Filipe A,Smollett K,Gu Q,Robertson N,Adams PD,Rai TS,Boutell C
Fri Mar 01 00:00:00 EST 2019
NAC1 Potentiates Cellular Antiviral Signaling by Bridging MAVS and TBK1.
Journal of immunology (Baltimore, Md. : 1950)
Xia Z,Xu G,Nie L,Liu L,Peng N,He Q,Zuo Q,Zhou Y,Cao Z,Liu S,Zhu Y
Thu Aug 15 00:00:00 EDT 2019
Avian Flavivirus Infection of Monocytes/Macrophages by Extensive Subversion of Host Antiviral Innate Immune Responses.
Journal of virology
Ma Y,Liang Y,Wang N,Cui L,Chen Z,Wu H,Zhu C,Wang Z,Liu S,Li H
Fri Nov 15 00:00:00 EST 2019
Repeated semen exposure decreases cervicovaginal SIVmac251 infection in rhesus macaques.
Nature communications
Abdulhaqq SA,Martinez M,Kang G,Rodriguez IV,Nichols SM,Beaumont D,Joseph J,Azzoni L,Yin X,Wise M,Weiner D,Liu Q,Foulkes A,Münch J,Kirchhoff F,Coutifaris C,Tomaras GD,Sariol C,Marx PA,Li Q,Kraiselburd EN,Montaner LJ
Wed Aug 21 00:00:00 EDT 2019
TBK1: A key regulator and potential treatment target for interferon positive Sjögren's syndrome, systemic lupus erythematosus and systemic sclerosis.
Journal of autoimmunity
Bodewes ILA,Huijser E,van Helden-Meeuwsen CG,Tas L,Huizinga R,Dalm VASH,van Hagen PM,Groot N,Kamphuis S,van Daele PLA,Versnel MA
Sun Jul 01 00:00:00 EDT 2018
TBK1: A key regulator and potential treatment target for interferon positive Sjögren's syndrome, systemic lupus erythematosus and systemic sclerosis.
Journal of autoimmunity
Bodewes ILA,Huijser E,van Helden-Meeuwsen CG,Tas L,Huizinga R,Dalm VASH,van Hagen PM,Groot N,Kamphuis S,van Daele PLA,Versnel MA
Sun Jul 01 00:00:00 EDT 2018
Interferon-alpha enhances the antitumour activity of EGFR-targeted therapies by upregulating RIG-I in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Jin S,Yang W,Zhou G,Zhao M,Fang S,Zhang Z,Hu J
Tue Feb 20 00:00:00 EST 2018
Interferon-alpha enhances the antitumour activity of EGFR-targeted therapies by upregulating RIG-I in head and neck squamous cell carcinoma.
British journal of cancer
Ma H,Jin S,Yang W,Zhou G,Zhao M,Fang S,Zhang Z,Hu J
Tue Feb 20 00:00:00 EST 2018
Phospholipid scramblase 1 interacts with influenza A virus NP, impairing its nuclear import and thereby suppressing virus replication.
PLoS pathogens
Luo W,Zhang J,Liang L,Wang G,Li Q,Zhu P,Zhou Y,Li J,Zhao Y,Sun N,Huang S,Zhou C,Chang Y,Cui P,Chen P,Jiang Y,Deng G,Bu Z,Li C,Jiang L,Chen H
Mon Jan 01 00:00:00 EST 2018
Phospholipid scramblase 1 interacts with influenza A virus NP, impairing its nuclear import and thereby suppressing virus replication.
PLoS pathogens
Luo W,Zhang J,Liang L,Wang G,Li Q,Zhu P,Zhou Y,Li J,Zhao Y,Sun N,Huang S,Zhou C,Chang Y,Cui P,Chen P,Jiang Y,Deng G,Bu Z,Li C,Jiang L,Chen H
Mon Jan 01 00:00:00 EST 2018
Distinct temporal roles for the promyelocytic leukaemia (PML) protein in the sequential regulation of intracellular host immunity to HSV-1 infection.
PLoS pathogens
Alandijany T,Roberts APE,Conn KL,Loney C,McFarlane S,Orr A,Boutell C
Mon Jan 01 00:00:00 EST 2018
Distinct temporal roles for the promyelocytic leukaemia (PML) protein in the sequential regulation of intracellular host immunity to HSV-1 infection.
PLoS pathogens
Alandijany T,Roberts APE,Conn KL,Loney C,McFarlane S,Orr A,Boutell C
Mon Jan 01 00:00:00 EST 2018
Bexarotene protects against neurotoxicity partially through a PPARγ-dependent mechanism in mice following traumatic brain injury.
Neurobiology of disease
He J,Liu H,Zhong J,Guo Z,Wu J,Zhang H,Huang Z,Jiang L,Li H,Zhang Z,Liu L,Wu Y,Qi L,Sun X,Cheng C
Sat Sep 01 00:00:00 EDT 2018
Bexarotene protects against neurotoxicity partially through a PPARγ-dependent mechanism in mice following traumatic brain injury.
Neurobiology of disease
He J,Liu H,Zhong J,Guo Z,Wu J,Zhang H,Huang Z,Jiang L,Li H,Zhang Z,Liu L,Wu Y,Qi L,Sun X,Cheng C
Sat Sep 01 00:00:00 EDT 2018
Bexarotene protects against neurotoxicity partially through a PPARγ-dependent mechanism in mice following traumatic brain injury.
Neurobiology of disease
He J,Liu H,Zhong J,Guo Z,Wu J,Zhang H,Huang Z,Jiang L,Li H,Zhang Z,Liu L,Wu Y,Qi L,Sun X,Cheng C
Sat Sep 01 00:00:00 EDT 2018
Bexarotene protects against neurotoxicity partially through a PPARγ-dependent mechanism in mice following traumatic brain injury.
Neurobiology of disease
He J,Liu H,Zhong J,Guo Z,Wu J,Zhang H,Huang Z,Jiang L,Li H,Zhang Z,Liu L,Wu Y,Qi L,Sun X,Cheng C
Sat Sep 01 00:00:00 EDT 2018
The large GTPase Mx1 binds Kif5B for cargo transport along microtubules.
Traffic (Copenhagen, Denmark)
Ringer K,Riehl J,Müller M,Dewes J,Hoff F,Jacob R
Sat Dec 01 00:00:00 EST 2018
The large GTPase Mx1 binds Kif5B for cargo transport along microtubules.
Traffic (Copenhagen, Denmark)
Ringer K,Riehl J,Müller M,Dewes J,Hoff F,Jacob R
Sat Dec 01 00:00:00 EST 2018
The large GTPase Mx1 binds Kif5B for cargo transport along microtubules.
Traffic (Copenhagen, Denmark)
Ringer K,Riehl J,Müller M,Dewes J,Hoff F,Jacob R
Sat Dec 01 00:00:00 EST 2018
The large GTPase Mx1 binds Kif5B for cargo transport along microtubules.
Traffic (Copenhagen, Denmark)
Ringer K,Riehl J,Müller M,Dewes J,Hoff F,Jacob R
Sat Dec 01 00:00:00 EST 2018
Type I IFN signature in childhood-onset systemic lupus erythematosus: a conspiracy of DNA- and RNA-sensing receptors?
Arthritis research & therapy
Wahadat MJ,Bodewes ILA,Maria NI,van Helden-Meeuwsen CG,van Dijk-Hummelman A,Steenwijk EC,Kamphuis S,Versnel MA
Wed Jan 10 00:00:00 EST 2018
Type I IFN signature in childhood-onset systemic lupus erythematosus: a conspiracy of DNA- and RNA-sensing receptors?
Arthritis research & therapy
Wahadat MJ,Bodewes ILA,Maria NI,van Helden-Meeuwsen CG,van Dijk-Hummelman A,Steenwijk EC,Kamphuis S,Versnel MA
Wed Jan 10 00:00:00 EST 2018
Characterization of Whooper Swan (Cygnus cygnus) Interferon α: Prokaryotic Expression, Biological Activities, and Physicochemical Characteristics.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Zhao H,Wang Y,Shao Y,Liu J,Li J,Zong H,Xing M
Mon Jan 01 00:00:00 EST 2018
Characterization of Whooper Swan (Cygnus cygnus) Interferon α: Prokaryotic Expression, Biological Activities, and Physicochemical Characteristics.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Zhao H,Wang Y,Shao Y,Liu J,Li J,Zong H,Xing M
Mon Jan 01 00:00:00 EST 2018
Myxovirus resistance protein A inhibits hepatitis C virus replication through JAK-STAT pathway activation.
Archives of virology
Wang H,Xin X,Wang M,Han L,Li J,Hao Y,Zheng C,Shen C
Fri Jun 01 00:00:00 EDT 2018
Myxovirus resistance protein A inhibits hepatitis C virus replication through JAK-STAT pathway activation.
Archives of virology
Wang H,Xin X,Wang M,Han L,Li J,Hao Y,Zheng C,Shen C
Fri Jun 01 00:00:00 EDT 2018
Alcohol attenuates anti-HCV function of IFN-λ1 through up-regulation of PLASy expression in human hepatic cells.
Journal of medical virology
Ran M,Huang J,Liang H,Jiang J,Liang B,Ning C,Zang N,Liao W,Liu H,Qin F,Yang Q,Ho W,Ye L,Chen H
Fri Jun 01 00:00:00 EDT 2018
Inducible Rubicon facilitates viral replication by antagonizing interferon production.
Cellular & molecular immunology
Wan Y,Cao W,Han T,Ren S,Feng J,Chen T,Wang J,Broering R,Lu M,Zhu Y
Sat Jul 01 00:00:00 EDT 2017
Inducible Rubicon facilitates viral replication by antagonizing interferon production.
Cellular & molecular immunology
Wan Y,Cao W,Han T,Ren S,Feng J,Chen T,Wang J,Broering R,Lu M,Zhu Y
Sat Jul 01 00:00:00 EDT 2017
Matrix Metalloproteinase 9 Facilitates Hepatitis B Virus Replication through Binding with Type I Interferon (IFN) Receptor 1 To Repress IFN/JAK/STAT Signaling.
Journal of virology
Chen J,Xu W,Chen Y,Xie X,Zhang Y,Ma C,Yang Q,Han Y,Zhu C,Xiong Y,Wu K,Liu F,Liu Y,Wu J
Sat Apr 15 00:00:00 EDT 2017
Matrix Metalloproteinase 9 Facilitates Hepatitis B Virus Replication through Binding with Type I Interferon (IFN) Receptor 1 To Repress IFN/JAK/STAT Signaling.
Journal of virology
Chen J,Xu W,Chen Y,Xie X,Zhang Y,Ma C,Yang Q,Han Y,Zhu C,Xiong Y,Wu K,Liu F,Liu Y,Wu J
Sat Apr 15 00:00:00 EDT 2017
Inducible TAP1 Negatively Regulates the Antiviral Innate Immune Response by Targeting the TAK1 Complex.
Journal of immunology (Baltimore, Md. : 1950)
Xia Z,Xu G,Yang X,Peng N,Zuo Q,Zhu S,Hao H,Liu S,Zhu Y
Mon May 01 00:00:00 EDT 2017
Inducible TAP1 Negatively Regulates the Antiviral Innate Immune Response by Targeting the TAK1 Complex.
Journal of immunology (Baltimore, Md. : 1950)
Xia Z,Xu G,Yang X,Peng N,Zuo Q,Zhu S,Hao H,Liu S,Zhu Y
Mon May 01 00:00:00 EDT 2017
Association of Increased Treg Cell Levels With Elevated Indoleamine 2,3-Dioxygenase Activity and an Imbalanced Kynurenine Pathway in Interferon-Positive Primary Sjögren's Syndrome.
Arthritis & rheumatology (Hoboken, N.J.)
Maria NI,van Helden-Meeuwsen CG,Brkic Z,Paulissen SM,Steenwijk EC,Dalm VA,van Daele PL,Martin van Hagen P,Kroese FG,van Roon JA,Harkin A,Dik WA,Drexhage HA,Lubberts E,Versnel MA
Fri Jul 01 00:00:00 EDT 2016
Association of Increased Treg Cell Levels With Elevated Indoleamine 2,3-Dioxygenase Activity and an Imbalanced Kynurenine Pathway in Interferon-Positive Primary Sjögren's Syndrome.
Arthritis & rheumatology (Hoboken, N.J.)
Maria NI,van Helden-Meeuwsen CG,Brkic Z,Paulissen SM,Steenwijk EC,Dalm VA,van Daele PL,Martin van Hagen P,Kroese FG,van Roon JA,Harkin A,Dik WA,Drexhage HA,Lubberts E,Versnel MA
Fri Jul 01 00:00:00 EDT 2016
Complex Interplay between HIV-1 Capsid and MX2-Independent Alpha Interferon-Induced Antiviral Factors.
Journal of virology
Bulli L,Apolonia L,Kutzner J,Pollpeter D,Goujon C,Herold N,Schwarz SM,Giernat Y,Keppler OT,Malim MH,Schaller T
Mon Aug 15 00:00:00 EDT 2016
Complex Interplay between HIV-1 Capsid and MX2-Independent Alpha Interferon-Induced Antiviral Factors.
Journal of virology
Bulli L,Apolonia L,Kutzner J,Pollpeter D,Goujon C,Herold N,Schwarz SM,Giernat Y,Keppler OT,Malim MH,Schaller T
Mon Aug 15 00:00:00 EDT 2016
MxA Mediates SUMO-Induced Resistance to Vesicular Stomatitis Virus.
Journal of virology
Maarifi G,Hannoun Z,Geoffroy MC,El Asmi F,Zarrouk K,Nisole S,Blondel D,Chelbi-Alix MK
Fri Jul 15 00:00:00 EDT 2016
MxA Mediates SUMO-Induced Resistance to Vesicular Stomatitis Virus.
Journal of virology
Maarifi G,Hannoun Z,Geoffroy MC,El Asmi F,Zarrouk K,Nisole S,Blondel D,Chelbi-Alix MK
Fri Jul 15 00:00:00 EDT 2016
MxA Mediates SUMO-Induced Resistance to Vesicular Stomatitis Virus.
Journal of virology
Maarifi G,Hannoun Z,Geoffroy MC,El Asmi F,Zarrouk K,Nisole S,Blondel D,Chelbi-Alix MK
Fri Jul 15 00:00:00 EDT 2016
MxA Mediates SUMO-Induced Resistance to Vesicular Stomatitis Virus.
Journal of virology
Maarifi G,Hannoun Z,Geoffroy MC,El Asmi F,Zarrouk K,Nisole S,Blondel D,Chelbi-Alix MK
Fri Jul 15 00:00:00 EDT 2016
STAT1 and NF-κB Inhibitors Diminish Basal Interferon-Stimulated Gene Expression and Improve the Productive Infection of Oncolytic HSV in MPNST Cells.
Molecular cancer research : MCR
Jackson JD,Markert JM,Li L,Carroll SL,Cassady KA
Sun May 01 00:00:00 EDT 2016
STAT1 and NF-κB Inhibitors Diminish Basal Interferon-Stimulated Gene Expression and Improve the Productive Infection of Oncolytic HSV in MPNST Cells.
Molecular cancer research : MCR
Jackson JD,Markert JM,Li L,Carroll SL,Cassady KA
Sun May 01 00:00:00 EDT 2016
Soluble IL-6 Receptor and IL-27 Subunit p28 Protein Complex Mediate the Antiviral Response through the Type III IFN Pathway.
Journal of immunology (Baltimore, Md. : 1950)
Yang X,Hao H,Xia Z,Xu G,Cao Z,Chen X,Liu S,Zhu Y
Thu Sep 15 00:00:00 EDT 2016
Soluble IL-6 Receptor and IL-27 Subunit p28 Protein Complex Mediate the Antiviral Response through the Type III IFN Pathway.
Journal of immunology (Baltimore, Md. : 1950)
Yang X,Hao H,Xia Z,Xu G,Cao Z,Chen X,Liu S,Zhu Y
Thu Sep 15 00:00:00 EDT 2016
Comparative Proteome Analysis of Porcine Jejunum Tissues in Response to a Virulent Strain of Porcine Epidemic Diarrhea Virus and Its Attenuated Strain.
Viruses
Li Z,Chen F,Ye S,Guo X,Muhanmmad Memon A,Wu M,He Q
Tue Nov 29 00:00:00 EST 2016
Comparative Proteome Analysis of Porcine Jejunum Tissues in Response to a Virulent Strain of Porcine Epidemic Diarrhea Virus and Its Attenuated Strain.
Viruses
Li Z,Chen F,Ye S,Guo X,Muhanmmad Memon A,Wu M,He Q
Tue Nov 29 00:00:00 EST 2016
Human hepatitis B virus surface and e antigens inhibit major vault protein signaling in interferon induction pathways.
Journal of hepatology
Liu S,Peng N,Xie J,Hao Q,Zhang M,Zhang Y,Xia Z,Xu G,Zhao F,Wang Q,Han T,Zhu Y
Fri May 01 00:00:00 EDT 2015
Human hepatitis B virus surface and e antigens inhibit major vault protein signaling in interferon induction pathways.
Journal of hepatology
Liu S,Peng N,Xie J,Hao Q,Zhang M,Zhang Y,Xia Z,Xu G,Zhao F,Wang Q,Han T,Zhu Y
Fri May 01 00:00:00 EDT 2015
Peptidylarginine deiminase inhibition disrupts NET formation and protects against kidney, skin and vascular disease in lupus-prone MRL/lpr mice.
Annals of the rheumatic diseases
Knight JS,Subramanian V,O'Dell AA,Yalavarthi S,Zhao W,Smith CK,Hodgin JB,Thompson PR,Kaplan MJ
Tue Dec 01 00:00:00 EST 2015
Peptidylarginine deiminase inhibition disrupts NET formation and protects against kidney, skin and vascular disease in lupus-prone MRL/lpr mice.
Annals of the rheumatic diseases
Knight JS,Subramanian V,O'Dell AA,Yalavarthi S,Zhao W,Smith CK,Hodgin JB,Thompson PR,Kaplan MJ
Tue Dec 01 00:00:00 EST 2015
Interferon inducible antiviral MxA is inversely associated with prostate cancer and regulates cell cycle, invasion and Docetaxel induced apoptosis.
The Prostate
Brown SG,Knowell AE,Hunt A,Patel D,Bhosle S,Chaudhary J
Sun Feb 15 00:00:00 EST 2015
Interferon inducible antiviral MxA is inversely associated with prostate cancer and regulates cell cycle, invasion and Docetaxel induced apoptosis.
The Prostate
Brown SG,Knowell AE,Hunt A,Patel D,Bhosle S,Chaudhary J
Sun Feb 15 00:00:00 EST 2015
Interferon inducible antiviral MxA is inversely associated with prostate cancer and regulates cell cycle, invasion and Docetaxel induced apoptosis.
The Prostate
Brown SG,Knowell AE,Hunt A,Patel D,Bhosle S,Chaudhary J
Sun Feb 15 00:00:00 EST 2015
Interferon inducible antiviral MxA is inversely associated with prostate cancer and regulates cell cycle, invasion and Docetaxel induced apoptosis.
The Prostate
Brown SG,Knowell AE,Hunt A,Patel D,Bhosle S,Chaudhary J
Sun Feb 15 00:00:00 EST 2015
Interferon inducible antiviral MxA is inversely associated with prostate cancer and regulates cell cycle, invasion and Docetaxel induced apoptosis.
The Prostate
Brown SG,Knowell AE,Hunt A,Patel D,Bhosle S,Chaudhary J
Sun Feb 15 00:00:00 EST 2015
Hepatitis B virus hijacks CTHRC1 to evade host immunity and maintain replication.
Journal of molecular cell biology
Bai L,Zhang W,Tan L,Yang H,Ge M,Zhu C,Zhang R,Cao Y,Chen J,Luo Z,Ho W,Liu F,Wu K,Wu J
Tue Dec 01 00:00:00 EST 2015
NRAV, a long noncoding RNA, modulates antiviral responses through suppression of interferon-stimulated gene transcription.
Cell host & microbe
Ouyang J,Zhu X,Chen Y,Wei H,Chen Q,Chi X,Qi B,Zhang L,Zhao Y,Gao GF,Wang G,Chen JL
Wed Nov 12 00:00:00 EST 2014
NRAV, a long noncoding RNA, modulates antiviral responses through suppression of interferon-stimulated gene transcription.
Cell host & microbe
Ouyang J,Zhu X,Chen Y,Wei H,Chen Q,Chi X,Qi B,Zhang L,Zhao Y,Gao GF,Wang G,Chen JL
Wed Nov 12 00:00:00 EST 2014
MxA as a clinically applicable biomarker for identifying systemic interferon type I in primary Sjogren's syndrome.
Annals of the rheumatic diseases
Maria NI,Brkic Z,Waris M,van Helden-Meeuwsen CG,Heezen K,van de Merwe JP,van Daele PL,Dalm VA,Drexhage HA,Versnel MA
Sun Jun 01 00:00:00 EDT 2014
MxA as a clinically applicable biomarker for identifying systemic interferon type I in primary Sjogren's syndrome.
Annals of the rheumatic diseases
Maria NI,Brkic Z,Waris M,van Helden-Meeuwsen CG,Heezen K,van de Merwe JP,van Daele PL,Dalm VA,Drexhage HA,Versnel MA
Sun Jun 01 00:00:00 EDT 2014
Epicutaneous application of toll-like receptor 7 agonists leads to systemic autoimmunity in wild-type mice: a new model of systemic Lupus erythematosus.
Arthritis & rheumatology (Hoboken, N.J.)
Yokogawa M,Takaishi M,Nakajima K,Kamijima R,Fujimoto C,Kataoka S,Terada Y,Sano S
Sat Mar 01 00:00:00 EST 2014
Epicutaneous application of toll-like receptor 7 agonists leads to systemic autoimmunity in wild-type mice: a new model of systemic Lupus erythematosus.
Arthritis & rheumatology (Hoboken, N.J.)
Yokogawa M,Takaishi M,Nakajima K,Kamijima R,Fujimoto C,Kataoka S,Terada Y,Sano S
Sat Mar 01 00:00:00 EST 2014
Interferon induction of IFITM proteins promotes infection by human coronavirus OC43.
Proceedings of the National Academy of Sciences of the United States of America
Zhao X,Guo F,Liu F,Cuconati A,Chang J,Block TM,Guo JT
Tue May 06 00:00:00 EDT 2014
Interferon induction of IFITM proteins promotes infection by human coronavirus OC43.
Proceedings of the National Academy of Sciences of the United States of America
Zhao X,Guo F,Liu F,Cuconati A,Chang J,Block TM,Guo JT
Tue May 06 00:00:00 EDT 2014
Influenza A virus-induced degradation of eukaryotic translation initiation factor 4B contributes to viral replication by suppressing IFITM3 protein expression.
Journal of virology
Wang S,Chi X,Wei H,Chen Y,Chen Z,Huang S,Chen JL
Fri Aug 01 00:00:00 EDT 2014
Influenza A virus-induced degradation of eukaryotic translation initiation factor 4B contributes to viral replication by suppressing IFITM3 protein expression.
Journal of virology
Wang S,Chi X,Wei H,Chen Y,Chen Z,Huang S,Chen JL
Fri Aug 01 00:00:00 EDT 2014
The large GTPase Mx1 is involved in apical transport in MDCK cells.
Traffic (Copenhagen, Denmark)
Hoff F,Greb C,Hollmann C,Hönig E,Jacob R
Mon Sep 01 00:00:00 EDT 2014
The large GTPase Mx1 is involved in apical transport in MDCK cells.
Traffic (Copenhagen, Denmark)
Hoff F,Greb C,Hollmann C,Hönig E,Jacob R
Mon Sep 01 00:00:00 EDT 2014
IL-27, a cytokine, and IFN-λ1, a type III IFN, are coordinated to regulate virus replication through type I IFN.
Journal of immunology (Baltimore, Md. : 1950)
Cao Y,Zhang R,Zhang W,Zhu C,Yu Y,Song Y,Wang Q,Bai L,Liu Y,Wu K,Wu J
Wed Jan 15 00:00:00 EST 2014
IL-27, a cytokine, and IFN-λ1, a type III IFN, are coordinated to regulate virus replication through type I IFN.
Journal of immunology (Baltimore, Md. : 1950)
Cao Y,Zhang R,Zhang W,Zhu C,Yu Y,Song Y,Wang Q,Bai L,Liu Y,Wu K,Wu J
Wed Jan 15 00:00:00 EST 2014
Quantitative proteomic analysis reveals that transmissible gastroenteritis virus activates the JAK-STAT1 signaling pathway.
Journal of proteome research
An K,Fang L,Luo R,Wang D,Xie L,Yang J,Chen H,Xiao S
Fri Dec 05 00:00:00 EST 2014
Quantitative proteomic analysis reveals that transmissible gastroenteritis virus activates the JAK-STAT1 signaling pathway.
Journal of proteome research
An K,Fang L,Luo R,Wang D,Xie L,Yang J,Chen H,Xiao S
Fri Dec 05 00:00:00 EST 2014
Serial cervicovaginal exposures with replication-deficient SIVsm induce higher dendritic cell (pDC) and CD4+ T-cell infiltrates not associated with prevention but a more severe SIVmac251 infection of rhesus macaques.
Journal of acquired immune deficiency syndromes (1999)
Abdulhaqq SA,Martinez MI,Kang G,Foulkes AS,Rodriguez IV,Nichols SM,Hunter M,Sariol CA,Ruiz LA,Ross BN,Yin X,Speicher DW,Haase AT,Marx PA,Li Q,Kraiselburd EN,Montaner LJ
Tue Apr 01 00:00:00 EDT 2014
Serial cervicovaginal exposures with replication-deficient SIVsm induce higher dendritic cell (pDC) and CD4+ T-cell infiltrates not associated with prevention but a more severe SIVmac251 infection of rhesus macaques.
Journal of acquired immune deficiency syndromes (1999)
Abdulhaqq SA,Martinez MI,Kang G,Foulkes AS,Rodriguez IV,Nichols SM,Hunter M,Sariol CA,Ruiz LA,Ross BN,Yin X,Speicher DW,Haase AT,Marx PA,Li Q,Kraiselburd EN,Montaner LJ
Tue Apr 01 00:00:00 EDT 2014
Vpx rescue of HIV-1 from the antiviral state in mature dendritic cells is independent of the intracellular deoxynucleotide concentration.
Retrovirology
Reinhard C,Bottinelli D,Kim B,Luban J
Sat Feb 01 00:00:00 EST 2014
Vpx rescue of HIV-1 from the antiviral state in mature dendritic cells is independent of the intracellular deoxynucleotide concentration.
Retrovirology
Reinhard C,Bottinelli D,Kim B,Luban J
Sat Feb 01 00:00:00 EST 2014
IFITM3 restricts the morbidity and mortality associated with influenza.
Nature
Everitt AR,Clare S,Pertel T,John SP,Wash RS,Smith SE,Chin CR,Feeley EM,Sims JS,Adams DJ,Wise HM,Kane L,Goulding D,Digard P,Anttila V,Baillie JK,Walsh TS,Hume DA,Palotie A,Xue Y,Colonna V,Tyler-Smith C,Dunning J,Gordon SB,Smyth RL,Openshaw PJ,Dougan G,Brass AL,Kellam P
Sun Mar 25 00:00:00 EDT 2012
IFITM3 restricts the morbidity and mortality associated with influenza.
Nature
Everitt AR,Clare S,Pertel T,John SP,Wash RS,Smith SE,Chin CR,Feeley EM,Sims JS,Adams DJ,Wise HM,Kane L,Goulding D,Digard P,Anttila V,Baillie JK,Walsh TS,Hume DA,Palotie A,Xue Y,Colonna V,Tyler-Smith C,Dunning J,Gordon SB,Smyth RL,Openshaw PJ,Dougan G,Brass AL,Kellam P
Sun Mar 25 00:00:00 EDT 2012
MxA inhibits hepatitis B virus replication by interaction with hepatitis B core antigen.
Hepatology (Baltimore, Md.)
Li N,Zhang L,Chen L,Feng W,Xu Y,Chen F,Liu X,Chen Z,Liu W
Sat Sep 01 00:00:00 EDT 2012
MxA inhibits hepatitis B virus replication by interaction with hepatitis B core antigen.
Hepatology (Baltimore, Md.)
Li N,Zhang L,Chen L,Feng W,Xu Y,Chen F,Liu X,Chen Z,Liu W
Sat Sep 01 00:00:00 EDT 2012
IFITM3 inhibits influenza A virus infection by preventing cytosolic entry.
PLoS pathogens
Feeley EM,Sims JS,John SP,Chin CR,Pertel T,Chen LM,Gaiha GD,Ryan BJ,Donis RO,Elledge SJ,Brass AL
Sat Oct 01 00:00:00 EDT 2011
IFITM3 inhibits influenza A virus infection by preventing cytosolic entry.
PLoS pathogens
Feeley EM,Sims JS,John SP,Chin CR,Pertel T,Chen LM,Gaiha GD,Ryan BJ,Donis RO,Elledge SJ,Brass AL
Sat Oct 01 00:00:00 EDT 2011
IFITM3 inhibits influenza A virus infection by preventing cytosolic entry.
PLoS pathogens
Feeley EM,Sims JS,John SP,Chin CR,Pertel T,Chen LM,Gaiha GD,Ryan BJ,Donis RO,Elledge SJ,Brass AL
Sat Oct 01 00:00:00 EDT 2011
IFITM3 inhibits influenza A virus infection by preventing cytosolic entry.
PLoS pathogens
Feeley EM,Sims JS,John SP,Chin CR,Pertel T,Chen LM,Gaiha GD,Ryan BJ,Donis RO,Elledge SJ,Brass AL
Sat Oct 01 00:00:00 EDT 2011
Vpx rescues HIV-1 transduction of dendritic cells from the antiviral state established by type 1 interferon.
Retrovirology
Pertel T,Reinhard C,Luban J
Wed Jun 22 00:00:00 EDT 2011
Vpx rescues HIV-1 transduction of dendritic cells from the antiviral state established by type 1 interferon.
Retrovirology
Pertel T,Reinhard C,Luban J
Wed Jun 22 00:00:00 EDT 2011
Rac1 is required for oncolytic NDV replication in human cancer cells and establishes a link between tumorigenesis and sensitivity to oncolytic virus.
Oncogene
Puhlmann J,Puehler F,Mumberg D,Boukamp P,Beier R
Thu Apr 15 00:00:00 EDT 2010
Rac1 is required for oncolytic NDV replication in human cancer cells and establishes a link between tumorigenesis and sensitivity to oncolytic virus.
Oncogene
Puhlmann J,Puehler F,Mumberg D,Boukamp P,Beier R
Thu Apr 15 00:00:00 EDT 2010
Identification of five interferon-induced cellular proteins that inhibit west nile virus and dengue virus infections.
Journal of virology
Jiang D,Weidner JM,Qing M,Pan XB,Guo H,Xu C,Zhang X,Birk A,Chang J,Shi PY,Block TM,Guo JT
Sun Aug 01 00:00:00 EDT 2010
Identification of five interferon-induced cellular proteins that inhibit west nile virus and dengue virus infections.
Journal of virology
Jiang D,Weidner JM,Qing M,Pan XB,Guo H,Xu C,Zhang X,Birk A,Chang J,Shi PY,Block TM,Guo JT
Sun Aug 01 00:00:00 EDT 2010