Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
Immunogen sequence: NKCQYCRFQK CLAVGMSHNA IRFGRMPQAE KEKLLAEISS DIDQLNPESA DLRALAKHLY DSYIKSFPLT KAKARAILTG KTTDKSPFVI YDMNSLMMGE DKIKFKHITP LQEQSKEVAI RIFQGCQFRS VEAVQEITEY AKSIPGFVNL DLNDQVTLLK YGVHEIIYTM LASLMNKDGV LISEGQGFMT REFLKSLRKP FGDFMEPKFE FAVKFNALEL DDSDLAIFIA VIILSGDRPG LLNVKPIEDI QDNLLQALEL QLKLNHPESS QLFAKLLQKM TDLRQIVTEH VQLLQVIKKT ETDMSLHPLL QEIYKDLY
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HUMPPARG; Nuclear receptor subfamily 1 group C member 3; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; PPAR gamma; PPAR-gamma; PPARy
Gene Aliases: CIMT1; GLM1; NR1C3; PPAR-gamma; PPAR-gamma2; PPARG; PPARG1; PPARG2; PPARgamma; PPARgamma2
UniProt ID: (Human) P37231, (Mouse) P37238, (Rat) O88275
Entrez Gene ID: (Human) 5468, (Mouse) 19016, (Rat) 25664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support