Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
ZCCHC17 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cytosolic large ribosomal subunit; nucleolus; nucleic acid binding; zinc ion binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hpNO40s; nucleolar protein 40; Nucleolar protein of 40 kDa; Pnn-interacting nucleolar protein; pNO40; PS1D protein; putative S1 RNA binding domain protein; Putative S1 RNA-binding domain protein; Zinc finger CCHC domain-containing protein 17; zinc finger, CCHC domain containing 17
Gene Aliases: 2810055E05Rik; HSPC243; HSPC251; LDC4; pNO40; PS1D; RGD1565267; ZCCHC17
UniProt ID: (Human) Q9NP64, (Mouse) Q9ESX4
Entrez Gene ID: (Human) 51538, (Mouse) 619605, (Rat) 500555
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support