Product References
Extracellular HSP90 promotes differentiation of lens epithelial cells to fiber cells by activating LRP1-YAP-PROX1 axis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li J,Yu J,Huang W,Sang F,Li J,Ren Y,Huang H,Wang M,Li K,Zhang J,Li H,Cui X,Zhang J,Hu M,Yuan F,Guo W,Zhang F,Mu H,Hu Y
Wed Feb 01 00:00:00 EST 2023
Herpetin Promotes Bone Marrow Mesenchymal Stem Cells to Alleviate Carbon Tetrachloride-Induced Acute Liver Injury in Mice.
Molecules (Basel, Switzerland)
Ding Y,Tan R,Gu J,Gong P
Mon May 01 00:00:00 EDT 2023
Insulin ameliorates dim blue light at night-induced apoptosis in hippocampal neurons via the IR/IRS1/AKT/GSK3β/β-catenin signaling pathway.
Ecotoxicology and environmental safety
Liu Q,Wang Z,Cao J,Dong Y,Chen Y
Sun Jan 15 00:00:00 EST 2023
Liraglutide Attenuates Hepatic Oxidative Stress, Inflammation, and Apoptosis in Streptozotocin-Induced Diabetic Mice by Modulating the Wnt/β-Catenin Signaling Pathway.
Mediators of inflammation
Yu J,Zhao Y,Xu L,Li W,Zhang H,Ping F,Li Y
Fri Feb 10 00:00:00 EST 2023
Hordenine Activated Dermal Papilla Cells and Promoted Hair Regrowth by Activating Wnt Signaling Pathway.
Nutrients
Wang C,Zang K,Tang Z,Yang T,Ye X,Dang Y
Mon Jan 30 00:00:00 EST 2023
Hordenine Activated Dermal Papilla Cells and Promoted Hair Regrowth by Activating Wnt Signaling Pathway.
Nutrients
Wang C,Zang K,Tang Z,Yang T,Ye X,Dang Y
Mon Jan 30 00:00:00 EST 2023
METTL14 Regulates PLAGL2/β-Catenin Signaling Axis to Promote the Development of Nonsmall Cell Lung Cancer.
Journal of oncology
Zhou Q,Lai X,Gao Y,Chen Q,Xu Y,Liu Y
Tue Mar 07 00:00:00 EST 2023
VASN promotes colorectal cancer progression by activating the YAP/TAZ and AKT signaling pathways via YAP.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Liang W,Zuo J,Liu M,Su Y,Guo B,Hou J,Xing Q,Peng Y,Fang L,Cao Y,Shan J,Sun R,Zhao J,Wang J
Sun Jan 01 00:00:00 EST 2023
FUT2 promotes the tumorigenicity and metastasis of colorectal cancer cells via the Wnt/β‑catenin pathway.
International journal of oncology
Liu P,Liu J,Ding M,Liu Y,Zhang Y,Chen X,Zhou Z
Wed Mar 01 00:00:00 EST 2023
FUT2 promotes the tumorigenicity and metastasis of colorectal cancer cells via the Wnt/β‑catenin pathway.
International journal of oncology
Liu P,Liu J,Ding M,Liu Y,Zhang Y,Chen X,Zhou Z
Wed Mar 01 00:00:00 EST 2023
B-lymphoid tyrosine kinase-mediated FAM83A phosphorylation elevates pancreatic tumorigenesis through interacting with β-catenin.
Signal transduction and targeted therapy
Zhou C,Zhu X,Liu N,Dong X,Zhang X,Huang H,Tang Y,Liu S,Hu M,Wang M,Deng X,Li S,Zhang R,Huang Y,Lyu H,Xiao S,Luo S,Ali DW,Michalak M,Chen XZ,Wang Z,Tang J
Fri Feb 17 00:00:00 EST 2023
B-lymphoid tyrosine kinase-mediated FAM83A phosphorylation elevates pancreatic tumorigenesis through interacting with β-catenin.
Signal transduction and targeted therapy
Zhou C,Zhu X,Liu N,Dong X,Zhang X,Huang H,Tang Y,Liu S,Hu M,Wang M,Deng X,Li S,Zhang R,Huang Y,Lyu H,Xiao S,Luo S,Ali DW,Michalak M,Chen XZ,Wang Z,Tang J
Fri Feb 17 00:00:00 EST 2023
B-lymphoid tyrosine kinase-mediated FAM83A phosphorylation elevates pancreatic tumorigenesis through interacting with β-catenin.
Signal transduction and targeted therapy
Zhou C,Zhu X,Liu N,Dong X,Zhang X,Huang H,Tang Y,Liu S,Hu M,Wang M,Deng X,Li S,Zhang R,Huang Y,Lyu H,Xiao S,Luo S,Ali DW,Michalak M,Chen XZ,Wang Z,Tang J
Fri Feb 17 00:00:00 EST 2023
ADAMDEC1 induces EMT and promotes colorectal cancer cells metastasis by enhancing Wnt/β-catenin signaling via negative modulation of GSK-3β.
Experimental cell research
Jia Y,Huang X,Shi H,Wang M,Chen J,Zhang H,Hou D,Jing H,Du J,Han H,Zhang J
Tue Aug 15 00:00:00 EDT 2023
ADAMDEC1 induces EMT and promotes colorectal cancer cells metastasis by enhancing Wnt/β-catenin signaling via negative modulation of GSK-3β.
Experimental cell research
Jia Y,Huang X,Shi H,Wang M,Chen J,Zhang H,Hou D,Jing H,Du J,Han H,Zhang J
Tue Aug 15 00:00:00 EDT 2023
Casein Kinase 1α Regulates Testosterone Synthesis and Testis Development in Adult Mice.
Endocrinology
Guo H,Zhang D,Zhou Y,Sun L,Li C,Luo X,Liu J,Cui S
Mon Mar 13 00:00:00 EDT 2023
Clinicopathological features and immunophenotype of Silva pattern system in endocervical adenocarcinoma.
International journal of experimental pathology
Zeng C,Wu JK,Lu X
Thu Jun 01 00:00:00 EDT 2023
Loss of CBX2 causes genomic instability and Wnt activation in high grade serous ovarian carcinoma cells.
Molecular carcinogenesis
Ma Y,Liu L,Wei Z,Zhu M,Huang L,Wang S,Yi X,Ying F,Zhao S,Cai J,Wang Z,Sun S
Sat Apr 01 00:00:00 EDT 2023
Novel biphasic mechanism of the canonical Wnt signalling component PYGO2 promotes cardiomyocyte differentiation from hUC-MSCs.
Cell and tissue research
Shi Y,Qin B,Fan X,Li Y,Wang Y,Yuan W,Jiang Z,Zhu P,Chen J,Chen Y,Li F,Wan Y,Wu X,Zhuang J
Sat Jul 01 00:00:00 EDT 2023
Novel biphasic mechanism of the canonical Wnt signalling component PYGO2 promotes cardiomyocyte differentiation from hUC-MSCs.
Cell and tissue research
Shi Y,Qin B,Fan X,Li Y,Wang Y,Yuan W,Jiang Z,Zhu P,Chen J,Chen Y,Li F,Wan Y,Wu X,Zhuang J
Sat Jul 01 00:00:00 EDT 2023
Tax1 binding protein 3 regulates osteogenic and adipogenic differentiation through inactivating Wnt/β-catenin signalling.
Journal of cellular and molecular medicine
Wang Y,Gan Y,Dong Y,Zhou J,Zhu E,Yuan H,Li X,Wang B
Sat Apr 01 00:00:00 EDT 2023
Tax1 binding protein 3 regulates osteogenic and adipogenic differentiation through inactivating Wnt/β-catenin signalling.
Journal of cellular and molecular medicine
Wang Y,Gan Y,Dong Y,Zhou J,Zhu E,Yuan H,Li X,Wang B
Sat Apr 01 00:00:00 EDT 2023
LINC00886 Negatively Regulates Malignancy in Anaplastic Thyroid Cancer.
Endocrinology
Ma B,Luo Y,Xu W,Han L,Liu W,Liao T,Yang Y,Wang Y
Sat Feb 11 00:00:00 EST 2023
DSTN Hypomethylation Promotes Radiotherapy Resistance of Rectal Cancer by Activating the Wnt/β-Catenin Signaling Pathway.
International journal of radiation oncology, biology, physics
Wen R,Zhou L,Jiang S,Fan H,Zheng K,Yu Y,Gao X,Hao L,Lou Z,Yu G,Yang F,Zhang W
Fri Sep 01 00:00:00 EDT 2023
Lnc-PPP2R1B Mediates the Alternative Splicing of PPP2R1B by Interacting and Stabilizing HNRNPLL and Promotes Osteogenesis of MSCs.
Stem cell reviews and reports
Peng S,He T,Liu Y,Zheng L,Zhong Y,Niu Z,Zhang M,Yang S
Tue Aug 01 00:00:00 EDT 2023
Prolonged exposure to the herbicide atrazine promotes kidney fibrosis by activating Wnt/β-catenin signaling in rats.
Environmental toxicology
Wang T,Huang X,Liu J,Liu W,Yang Z,He K,Chen J,Zhao L
Mon May 01 00:00:00 EDT 2023
Prolonged exposure to the herbicide atrazine promotes kidney fibrosis by activating Wnt/β-catenin signaling in rats.
Environmental toxicology
Wang T,Huang X,Liu J,Liu W,Yang Z,He K,Chen J,Zhao L
Mon May 01 00:00:00 EDT 2023
Barrier-penetrating liposome targeted delivery of basic fibroblast growth factor for spinal cord injury repair.
Materials today. Bio
Wu F,Wang P,Wei X,Yang Y,Al Mamun A,Zhang X,Zhu Y,Mo T,Zhang H,Jiang C,Hu J,Xiao J
Wed Feb 01 00:00:00 EST 2023
Barrier-penetrating liposome targeted delivery of basic fibroblast growth factor for spinal cord injury repair.
Materials today. Bio
Wu F,Wang P,Wei X,Yang Y,Al Mamun A,Zhang X,Zhu Y,Mo T,Zhang H,Jiang C,Hu J,Xiao J
Wed Feb 01 00:00:00 EST 2023
Barrier-penetrating liposome targeted delivery of basic fibroblast growth factor for spinal cord injury repair.
Materials today. Bio
Wu F,Wang P,Wei X,Yang Y,Al Mamun A,Zhang X,Zhu Y,Mo T,Zhang H,Jiang C,Hu J,Xiao J
Wed Feb 01 00:00:00 EST 2023
Barrier-penetrating liposome targeted delivery of basic fibroblast growth factor for spinal cord injury repair.
Materials today. Bio
Wu F,Wang P,Wei X,Yang Y,Al Mamun A,Zhang X,Zhu Y,Mo T,Zhang H,Jiang C,Hu J,Xiao J
Wed Feb 01 00:00:00 EST 2023
Non-muscle myosins control radial glial basal endfeet to mediate interneuron organization.
PLoS biology
D'Arcy BR,Lennox AL,Manso Musso C,Bracher A,Escobar-Tomlienovich C,Perez-Sanchez S,Silver DL
Wed Feb 01 00:00:00 EST 2023
Porcupine inhibitor CGX1321 alleviates heart failure with preserved ejection fraction in mice by blocking WNT signaling.
Acta pharmacologica Sinica
Wu H,Tang LX,Wang XM,Li LP,Chen XK,He YJ,Yang DZ,Shi Y,Shou JL,Zhang ZS,Wang L,Lu BJ,An SM,Zeng CY,Wang WE
Thu Jun 01 00:00:00 EDT 2023
Porcupine inhibitor CGX1321 alleviates heart failure with preserved ejection fraction in mice by blocking WNT signaling.
Acta pharmacologica Sinica
Wu H,Tang LX,Wang XM,Li LP,Chen XK,He YJ,Yang DZ,Shi Y,Shou JL,Zhang ZS,Wang L,Lu BJ,An SM,Zeng CY,Wang WE
Thu Jun 01 00:00:00 EDT 2023
PRMT2 promotes RCC tumorigenesis and metastasis via enhancing WNT5A transcriptional expression.
Cell death & disease
Li Z,Chen C,Yong H,Jiang L,Wang P,Meng S,Chu S,Li Z,Guo Q,Zheng J,Bai J,Li H
Fri May 12 00:00:00 EDT 2023
STMN2 overexpression promotes cell proliferation and EMT in pancreatic cancer mediated by WNT/β-catenin signaling.
Cancer gene therapy
Shao M,Wang L,Zhang Q,Wang T,Wang S
Wed Mar 01 00:00:00 EST 2023
STMN2 overexpression promotes cell proliferation and EMT in pancreatic cancer mediated by WNT/β-catenin signaling.
Cancer gene therapy
Shao M,Wang L,Zhang Q,Wang T,Wang S
Wed Mar 01 00:00:00 EST 2023
STMN2 overexpression promotes cell proliferation and EMT in pancreatic cancer mediated by WNT/β-catenin signaling.
Cancer gene therapy
Shao M,Wang L,Zhang Q,Wang T,Wang S
Wed Mar 01 00:00:00 EST 2023
STMN2 overexpression promotes cell proliferation and EMT in pancreatic cancer mediated by WNT/β-catenin signaling.
Cancer gene therapy
Shao M,Wang L,Zhang Q,Wang T,Wang S
Wed Mar 01 00:00:00 EST 2023
STMN2 overexpression promotes cell proliferation and EMT in pancreatic cancer mediated by WNT/β-catenin signaling.
Cancer gene therapy
Shao M,Wang L,Zhang Q,Wang T,Wang S
Wed Mar 01 00:00:00 EST 2023
STMN2 overexpression promotes cell proliferation and EMT in pancreatic cancer mediated by WNT/β-catenin signaling.
Cancer gene therapy
Shao M,Wang L,Zhang Q,Wang T,Wang S
Wed Mar 01 00:00:00 EST 2023
Vascular Endothelial Growth Factor A Contributes to Increased Mammalian Respiratory Epithelial Permeability Induced by Pasteurella multocida Infection.
Microbiology spectrum
Lin L,Yang J,Zhang D,Lv Q,Wang F,Liu P,Wang M,Shi C,Huang X,Liang W,Tan C,Wang X,Chen H,Wilson BA,Wu B,Peng Z
Tue Mar 14 00:00:00 EDT 2023
Crosstalk between 5-methylcytosine and N6-methyladenosine machinery defines disease progression, therapeutic response and pharmacogenomic landscape in hepatocellular carcinoma.
Molecular cancer
Tian Y,Xiao H,Yang Y,Zhang P,Yuan J,Zhang W,Chen L,Fan Y,Zhang J,Cheng H,Deng T,Yang L,Wang W,Chen G,Wang P,Gong P,Niu X,Zhang X
Tue Jan 10 00:00:00 EST 2023
RRP15 deficiency induces ribosome stress to inhibit colorectal cancer proliferation and metastasis via LZTS2-mediated β-catenin suppression.
Cell death & disease
Dong Z,Li J,Dai W,Yu D,Zhao Y,Liu S,Li X,Zhang Z,Zhang R,Liang X,Kong Q,Jin S,Jiang H,Jiang W,Ding C
Tue Feb 07 00:00:00 EST 2023
RRP15 deficiency induces ribosome stress to inhibit colorectal cancer proliferation and metastasis via LZTS2-mediated β-catenin suppression.
Cell death & disease
Dong Z,Li J,Dai W,Yu D,Zhao Y,Liu S,Li X,Zhang Z,Zhang R,Liang X,Kong Q,Jin S,Jiang H,Jiang W,Ding C
Tue Feb 07 00:00:00 EST 2023
Disulfiram suppresses epithelial-mesenchymal transition (EMT), migration and invasion in cervical cancer through the HSP90A/NDRG1 pathway.
Cellular signalling
Zheng P,Wu Y,Wang Y,Hu F
Fri Sep 01 00:00:00 EDT 2023
Anti-Postmenopausal osteoporosis effects of Isopsoralen: A bioinformatics-integrated experimental study.
Phytotherapy research : PTR
Li H,Wang C,Jin Y,Cai Y,Yao J,Meng Q,Wu J,Wang H,Sun H,Liu M
Sun Jan 01 00:00:00 EST 2023
lncRNA MEG3 Promotes Osteogenic Differentiation of Tendon Stem Cells Via the miR-129-5p/TCF4/β-Catenin Axis and thus Contributes to Trauma-Induced Heterotopic Ossification.
Stem cell reviews and reports
Liu H,Sun Z,Luo G,Hu Y,Ruan H,Tu B,Li J,Fan C
Sun Oct 01 00:00:00 EDT 2023
lncRNA MEG3 Promotes Osteogenic Differentiation of Tendon Stem Cells Via the miR-129-5p/TCF4/β-Catenin Axis and thus Contributes to Trauma-Induced Heterotopic Ossification.
Stem cell reviews and reports
Liu H,Sun Z,Luo G,Hu Y,Ruan H,Tu B,Li J,Fan C
Sun Oct 01 00:00:00 EDT 2023
TREM2 as an independent predictor of poor prognosis promotes the migration via the PI3K/AKT axis in prostate cancer.
American journal of translational research
Gao HT,Yang Z,Sun H,Zhang Y,Wang Z,Liu WY,Wen HZ,Qu CB,Wang XL
Wed Mar 15 00:00:00 EDT 2023
Evaluation of toxicological effects of bisphenol S with an in vitro human bone marrow mesenchymal stem cell: Implications for bone health.
Toxicology
Li M,Li T,Yin J,Xie C,Zhu J
Sun Jan 15 00:00:00 EST 2023
Evaluation of toxicological effects of bisphenol S with an in vitro human bone marrow mesenchymal stem cell: Implications for bone health.
Toxicology
Li M,Li T,Yin J,Xie C,Zhu J
Sun Jan 15 00:00:00 EST 2023
eIF3a sustains non-small cell lung cancer stem cell-like properties by promoting YY1-mediated transcriptional activation of β-catenin.
Biochemical pharmacology
Zheng JY,Zhu T,Zhuo W,Mao XY,Yin JY,Li X,He YJ,Zhang W,Liu C,Liu ZQ
Sat Jul 01 00:00:00 EDT 2023
The oncogenic and immunological roles of histidine triad nucleotide-binding protein 1 in human cancers and their experimental validation in the MCF-7 cell line.
Annals of translational medicine
Wang X,Zhou M,Jiang L
Wed Feb 15 00:00:00 EST 2023
CRISPR/Cas9 based gene editing of Frizzled class receptor 6 (FZD6) reveals its role in depressive symptoms through disrupting Wnt/β-catenin signaling pathway.
Journal of advanced research
Han H,Xu M,Wang J,Li MD,Yang Z
Wed Jun 14 00:00:00 EDT 2023
Liver-specific overexpression of HKDC1 increases hepatocyte size and proliferative capacity.
Scientific reports
Pusec CM,Ilievski V,De Jesus A,Farooq Z,Zapater JL,Sweis N,Ismail H,Khan MW,Ardehali H,Cordoba-Chacon J,Layden BT
Wed May 17 00:00:00 EDT 2023
Environmentally relevant perinatal exposure to DBP accelerated spermatogenesis by promoting the glycolipid metabolism of Sertoli cells in male offspring mice.
Toxicology letters
Ma T,Xia Y,Wang B,Yang F,Ding J,Wu J,Han X,Wang J,Li D
Wed Mar 15 00:00:00 EDT 2023
BMSC-derived extracellular vesicles promoted osteogenesis via Axin2 inhibition by delivering MiR-16-5p.
International immunopharmacology
Duan J,Li H,Wang C,Yao J,Jin Y,Zhao J,Zhang Y,Liu M,Sun H
Sat Jul 01 00:00:00 EDT 2023
Rhein Inhibited Ferroptosis and EMT to Attenuate Diabetic Nephropathy by Regulating the Rac1/NOX1/β-Catenin Axis.
Frontiers in bioscience (Landmark edition)
Xiong D,Hu W,Han X,Cai Y
Thu May 25 00:00:00 EDT 2023
Genetic modification of miR-34a enhances efficacy of transplanted human dental pulp stem cells after ischemic stroke.
Neural regeneration research
Wang J,He P,Tian Q,Luo Y,He Y,Liu C,Gong P,Guo Y,Ye Q,Li M
Fri Sep 01 00:00:00 EDT 2023
lncRNA HCG11 suppresses cell proliferation in hormone receptor-positive breast cancer via SRSF1/β-catenin.
Aging
Xie D,Li S,Wang X,Fang L
Wed Jan 04 00:00:00 EST 2023
lncRNA HCG11 suppresses cell proliferation in hormone receptor-positive breast cancer via SRSF1/β-catenin.
Aging
Xie D,Li S,Wang X,Fang L
Wed Jan 04 00:00:00 EST 2023
lncRNA HCG11 suppresses cell proliferation in hormone receptor-positive breast cancer via SRSF1/β-catenin.
Aging
Xie D,Li S,Wang X,Fang L
Wed Jan 04 00:00:00 EST 2023
lncRNA HCG11 suppresses cell proliferation in hormone receptor-positive breast cancer via SRSF1/β-catenin.
Aging
Xie D,Li S,Wang X,Fang L
Wed Jan 04 00:00:00 EST 2023
CircJag1 promotes apoptosis of ethylene thiourea-exposed anorectal malformations through sponging miR-137-3p by regulating Sox9 and suppressing Wnt/β-catenin pathway during the hindgut development of rat embryos.
Cell biology and toxicology
Li SY,Wang CY,Wei XG,Tang XB,Yuan ZW,Bai YZ
Tue Aug 01 00:00:00 EDT 2023
CircJag1 promotes apoptosis of ethylene thiourea-exposed anorectal malformations through sponging miR-137-3p by regulating Sox9 and suppressing Wnt/β-catenin pathway during the hindgut development of rat embryos.
Cell biology and toxicology
Li SY,Wang CY,Wei XG,Tang XB,Yuan ZW,Bai YZ
Tue Aug 01 00:00:00 EDT 2023
Enamel defects of Axenfeld-Rieger syndrome and the role of PITX2 in its pathogenesis.
Oral diseases
Yang Y,Zhu J,Chiba Y,Fukumoto S,Qin M,Wang X
Wed Nov 01 00:00:00 EDT 2023
ZIM1 Combined with Hydrogel Inhibits Senescence of Primary PαS Cells during In Vitro Expansion.
International journal of molecular sciences
Tian Y,Hu M,Liu X,Wang X,Lu D,Li Z,Liu Y,Zhang P,Zhou Y
Mon Jun 05 00:00:00 EDT 2023
Identification of the Main Chemical constituents and mechanism of Renshen Guben oral liquid against Renal Fibrosis.
Chinese medicine
Zhang J,Peng J,Zhang T,Jiang H,Qin Y,Chen H,Deng X,Ren J,Wang P,Xu H
Wed May 17 00:00:00 EDT 2023
Synergistic Effect of Magneto-Mechanical Bioengineered Stem Cells and Magnetic Field to Alleviate Osteoporosis.
ACS applied materials & interfaces
Yu C,Yang W,Yang L,Ye L,Sun R,Gu T,Ying X,Wang M,Tang R,Fan S,Yao S
Wed Apr 26 00:00:00 EDT 2023
Synergistic Effect of Magneto-Mechanical Bioengineered Stem Cells and Magnetic Field to Alleviate Osteoporosis.
ACS applied materials & interfaces
Yu C,Yang W,Yang L,Ye L,Sun R,Gu T,Ying X,Wang M,Tang R,Fan S,Yao S
Wed Apr 26 00:00:00 EDT 2023
Baicalin facilitates remyelination and suppresses neuroinflammation in rats with chronic cerebral hypoperfusion by activating Wnt/β-catenin and inhibiting NF-κB signaling.
Behavioural brain research
Xiao Y,Guan T,Yang X,Xu J,Zhang J,Qi Q,Teng Z,Dong Y,Gao Y,Li M,Meng N,Lv P
Tue Mar 28 00:00:00 EDT 2023
A2aR inhibits fibrosis and the EMT process in silicosis by regulating Wnt/β-catenin pathway.
Ecotoxicology and environmental safety
Tian Y,Xia J,Yang G,Li C,Qi Y,Dai K,Wu C,Guo Y,Yao W,Hao C
Sun Jan 01 00:00:00 EST 2023
A2aR inhibits fibrosis and the EMT process in silicosis by regulating Wnt/β-catenin pathway.
Ecotoxicology and environmental safety
Tian Y,Xia J,Yang G,Li C,Qi Y,Dai K,Wu C,Guo Y,Yao W,Hao C
Sun Jan 01 00:00:00 EST 2023
A2aR inhibits fibrosis and the EMT process in silicosis by regulating Wnt/β-catenin pathway.
Ecotoxicology and environmental safety
Tian Y,Xia J,Yang G,Li C,Qi Y,Dai K,Wu C,Guo Y,Yao W,Hao C
Sun Jan 01 00:00:00 EST 2023
A2aR inhibits fibrosis and the EMT process in silicosis by regulating Wnt/β-catenin pathway.
Ecotoxicology and environmental safety
Tian Y,Xia J,Yang G,Li C,Qi Y,Dai K,Wu C,Guo Y,Yao W,Hao C
Sun Jan 01 00:00:00 EST 2023
A2aR inhibits fibrosis and the EMT process in silicosis by regulating Wnt/β-catenin pathway.
Ecotoxicology and environmental safety
Tian Y,Xia J,Yang G,Li C,Qi Y,Dai K,Wu C,Guo Y,Yao W,Hao C
Sun Jan 01 00:00:00 EST 2023
A2aR inhibits fibrosis and the EMT process in silicosis by regulating Wnt/β-catenin pathway.
Ecotoxicology and environmental safety
Tian Y,Xia J,Yang G,Li C,Qi Y,Dai K,Wu C,Guo Y,Yao W,Hao C
Sun Jan 01 00:00:00 EST 2023
SOX9/NFIA promotes human ovarian cancer metastasis through the Wnt/β-catenin signaling pathway.
Pathology, research and practice
Lu R,Tang P,Zhang D,Lin S,Li H,Feng X,Sun M,Zhang H
Tue Aug 01 00:00:00 EDT 2023
Arsenic interferes with spermatogenesis involving Rictor/mTORC2-mediated blood-testis barrier disruption in mice.
Ecotoxicology and environmental safety
Li X,Wang W,Hou Y,Li G,Yi H,Cui S,Zhang J,He X,Zhao H,Yang Z,Qiu Y,Liu Z,Xie J
Thu Jun 01 00:00:00 EDT 2023
Arsenic interferes with spermatogenesis involving Rictor/mTORC2-mediated blood-testis barrier disruption in mice.
Ecotoxicology and environmental safety
Li X,Wang W,Hou Y,Li G,Yi H,Cui S,Zhang J,He X,Zhao H,Yang Z,Qiu Y,Liu Z,Xie J
Thu Jun 01 00:00:00 EDT 2023
Optogenetic Stimulation of mPFC Alleviates White Matter Injury-Related Cognitive Decline after Chronic Ischemia through Adaptive Myelination.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Deng S,Shu S,Zhai L,Xia S,Cao X,Li H,Bao X,Liu P,Xu Y
Wed Feb 01 00:00:00 EST 2023
Optogenetic Stimulation of mPFC Alleviates White Matter Injury-Related Cognitive Decline after Chronic Ischemia through Adaptive Myelination.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Deng S,Shu S,Zhai L,Xia S,Cao X,Li H,Bao X,Liu P,Xu Y
Wed Feb 01 00:00:00 EST 2023
CRISPR-Cas9-Mediated NPC1 Gene Deletion Enhances HEK 293 T Cell Adhesion by Regulating E-Cadherin.
Molecular biotechnology
Jia Z,Yang M,Zhao Y,Li X,Yang C,Qiao L,Li H,Du J,Lin J,Guan L
Wed Feb 01 00:00:00 EST 2023
UBP43 promotes epithelial ovarian carcinogenesis via activation of β-catenin signaling pathway.
Cell biology international
Xie H,Chen J,Ma C,Zhao J,Cui M
Tue Aug 01 00:00:00 EDT 2023
QiHuangYiShen Granules Modulate the Expression of LncRNA MALAT1 and Attenuate Epithelial-Mesenchymal Transition in Kidney of Diabetic Nephropathy Rats.
Evidence-based complementary and alternative medicine : eCAM
Yuan LS,Du H,Ren QY,Yang RL,Liu SW,Liu SY,Shi KF,Wang B,Meng XF,Li TX,Zhang N
Sat Feb 11 00:00:00 EST 2023
Ursolic Acid Suppresses Colorectal Cancer by Down-Regulation of Wnt/β-Catenin Signaling Pathway Activity.
Journal of agricultural and food chemistry
Zhao H,Tang S,Tao Q,Ming T,Lei J,Liang Y,Peng Y,Wang M,Liu M,Yang H,Ren S,Xu H
Wed Mar 08 00:00:00 EST 2023
Resveratrol suppresses lung cancer by targeting cancer stem-like cells and regulating tumor microenvironment.
The Journal of nutritional biochemistry
Xie C,Liang C,Wang R,Yi K,Zhou X,Li X,Chen Y,Miao D,Zhong C,Zhu J
Wed Feb 01 00:00:00 EST 2023
Resveratrol suppresses lung cancer by targeting cancer stem-like cells and regulating tumor microenvironment.
The Journal of nutritional biochemistry
Xie C,Liang C,Wang R,Yi K,Zhou X,Li X,Chen Y,Miao D,Zhong C,Zhu J
Wed Feb 01 00:00:00 EST 2023
Resveratrol suppresses lung cancer by targeting cancer stem-like cells and regulating tumor microenvironment.
The Journal of nutritional biochemistry
Xie C,Liang C,Wang R,Yi K,Zhou X,Li X,Chen Y,Miao D,Zhong C,Zhu J
Wed Feb 01 00:00:00 EST 2023
Resveratrol suppresses lung cancer by targeting cancer stem-like cells and regulating tumor microenvironment.
The Journal of nutritional biochemistry
Xie C,Liang C,Wang R,Yi K,Zhou X,Li X,Chen Y,Miao D,Zhong C,Zhu J
Wed Feb 01 00:00:00 EST 2023
N6-Methyladenosine-Mediated Up-Regulation of FZD10 Regulates Liver Cancer Stem Cells' Properties and Lenvatinib Resistance Through WNT/β-Catenin and Hippo Signaling Pathways.
Gastroenterology
Wang J,Yu H,Dong W,Zhang C,Hu M,Ma W,Jiang X,Li H,Yang P,Xiang D
Mon May 01 00:00:00 EDT 2023
RNF20 is required for male fertility through regulation of H2B ubiquitination in the Sertoli cells.
Cell & bioscience
Lai F,Wang H,Zhao X,Yang K,Cai L,Hu M,Lin L,Xia X,Li W,Cheng H,Zhou R
Thu Apr 06 00:00:00 EDT 2023
Cerium Oxide Nanoparticles Promote Osteoplastic Precursor Differentiation by Activating the Wnt Pathway.
Biological trace element research
Luo J,Zhu S,Tong Y,Zhang Y,Li Y,Cao L,Kong M,Luo M,Bi Q,Zhang Q
Wed Feb 01 00:00:00 EST 2023
Fam20c regulates the calpain proteolysis system through phosphorylating Calpasatatin to maintain cell homeostasis.
Journal of translational medicine
Liu X,Jiang L,Zhang W,Zhang J,Luan X,Zhan Y,Wang T,Da J,Liu L,Zhang S,Guo Y,Zhang K,Wang Z,Miao N,Xie X,Liu P,Li Y,Jin H,Zhang B
Tue Jun 27 00:00:00 EDT 2023
Fam20c regulates the calpain proteolysis system through phosphorylating Calpasatatin to maintain cell homeostasis.
Journal of translational medicine
Liu X,Jiang L,Zhang W,Zhang J,Luan X,Zhan Y,Wang T,Da J,Liu L,Zhang S,Guo Y,Zhang K,Wang Z,Miao N,Xie X,Liu P,Li Y,Jin H,Zhang B
Tue Jun 27 00:00:00 EDT 2023
Fam20c regulates the calpain proteolysis system through phosphorylating Calpasatatin to maintain cell homeostasis.
Journal of translational medicine
Liu X,Jiang L,Zhang W,Zhang J,Luan X,Zhan Y,Wang T,Da J,Liu L,Zhang S,Guo Y,Zhang K,Wang Z,Miao N,Xie X,Liu P,Li Y,Jin H,Zhang B
Tue Jun 27 00:00:00 EDT 2023
Chebulagic acid suppresses gastric cancer by inhibiting the AURKA/β-catenin/Wnt pathway.
Frontiers in pharmacology
Zhao J,Shi Y,Ma Y,Pan L,Wang Y,Yuan L,Dong J,Ying J
Tue Mar 21 00:00:00 EDT 2023
Onvansertib inhibits the proliferation and improves the cisplatin-resistance of lung adenocarcinoma via β-catenin/c-Myc signaling pathway.
American journal of cancer research
Wang R,Hou Y,Geng G,Zhu X,Wang Z,Cai W,Ye J,Zhao S,Mi Y,Jiang J
Sat Mar 11 00:00:00 EST 2023
Hastatoside attenuatescarbon tetrachloride-induced liver fibrosis by targeting glycogen synthase kinase-3β.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Du C,Dong J,Wang Q,Xu C,Feng S,Feng R,Lv X,Li J,Zhang L,Huang C,Ma T
Sun Jan 01 00:00:00 EST 2023
Hastatoside attenuatescarbon tetrachloride-induced liver fibrosis by targeting glycogen synthase kinase-3β.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Du C,Dong J,Wang Q,Xu C,Feng S,Feng R,Lv X,Li J,Zhang L,Huang C,Ma T
Sun Jan 01 00:00:00 EST 2023
Hastatoside attenuatescarbon tetrachloride-induced liver fibrosis by targeting glycogen synthase kinase-3β.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Du C,Dong J,Wang Q,Xu C,Feng S,Feng R,Lv X,Li J,Zhang L,Huang C,Ma T
Sun Jan 01 00:00:00 EST 2023
Hastatoside attenuatescarbon tetrachloride-induced liver fibrosis by targeting glycogen synthase kinase-3β.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Du C,Dong J,Wang Q,Xu C,Feng S,Feng R,Lv X,Li J,Zhang L,Huang C,Ma T
Sun Jan 01 00:00:00 EST 2023
Targeting CDK12 obviates the malignant phenotypes of colorectal cancer through the Wnt/β-catenin signaling pathway.
Experimental cell research
Liu S,Wu J,Lu X,Guo C,Zheng Q,Wang Y,Hu Q,Bian S,Luo L,Cheng Q,Liu Z,Dai W
Sat Jul 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Mitoquinone ameliorated airway inflammation by stabilizing β-catenin destruction complex in a steroid-insensitive asthma model.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang J,Chen Y,Peng X,Gong Z,Wang Y,Li Y,Xu M,Ma Y,Yu C,Cai S,Zhao W,Zhao H
Thu Jun 01 00:00:00 EDT 2023
Environmental exposure to triazole fungicide causes left-right asymmetry defects and contributes to abnormal heart development in zebrafish embryos by activating PPARγ-coupled Wnt/β-catenin signaling pathway.
The Science of the total environment
Wang Y,Ren Y,Ning X,Li G,Sang N
Fri Feb 10 00:00:00 EST 2023
Exosome-based bone-targeting drug delivery alleviates impaired osteoblastic bone formation and bone loss in inflammatory bowel diseases.
Cell reports. Medicine
Guo J,Wang F,Hu Y,Luo Y,Wei Y,Xu K,Zhang H,Liu H,Bo L,Lv S,Sheng S,Zhuang X,Zhang T,Xu C,Chen X,Su J
Tue Jan 17 00:00:00 EST 2023
PABPN1 regulates mRNA alternative polyadenylation to inhibit bladder cancer progression.
Cell & bioscience
Chen L,Dong W,Zhou M,Yang C,Xiong M,Kazobinka G,Chen Z,Xing Y,Hou T
Mon Mar 06 00:00:00 EST 2023
YAP/TEAD1 and β-catenin/LEF1 synergistically induce estrogen receptor α to promote osteogenic differentiation of bone marrow stromal cells.
MedComm
Wang P,Huang L,Yang F,Chen W,Bai D,Guo Y
Thu Jun 01 00:00:00 EDT 2023
Notch1 is a marker for in situ resting osteocytes in a 3-dimensional gel culture model.
Connective tissue research
Zhou YH,Zhu JY,Guo Y,Tang HN,Wang F,Iqbal J,Wu HX,Hu N,Xiao F,Wang T,Li L,Zhou HD
Fri Sep 01 00:00:00 EDT 2023
The Wnt/β-catenin pathway regulates inflammation and apoptosis in ventilator-induced lung injury.
Bioscience reports
Chen Z,He S,Lian S,Shen Y,Jiang W,Zhou L,Zhou L,Zhang X
Fri Mar 31 00:00:00 EDT 2023
β-catenin facilitates fowl adenovirus serotype 4 replication through enhancing virus-induced autophagy.
Veterinary microbiology
Wang T,Wang C,Han J,Hou X,Hu R,Chang W,Wang L,Qi X,Wang J
Sun Jan 01 00:00:00 EST 2023
β-catenin facilitates fowl adenovirus serotype 4 replication through enhancing virus-induced autophagy.
Veterinary microbiology
Wang T,Wang C,Han J,Hou X,Hu R,Chang W,Wang L,Qi X,Wang J
Sun Jan 01 00:00:00 EST 2023
AQP8 may affect glioma proliferation and growth by regulating GSK-3β phosphorylation and nuclear transport of β-catenin.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience
Cai Z,Shen Z,Zhao J,Zhang H,Guo Z,Xia Q,Liang H,Liu J,Tan L,Sheng H,Zhu S
Thu Jun 01 00:00:00 EDT 2023
AQP8 may affect glioma proliferation and growth by regulating GSK-3β phosphorylation and nuclear transport of β-catenin.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience
Cai Z,Shen Z,Zhao J,Zhang H,Guo Z,Xia Q,Liang H,Liu J,Tan L,Sheng H,Zhu S
Thu Jun 01 00:00:00 EDT 2023
Berberine plays a cardioprotective role by inhibiting macrophage Wnt5a/β-catenin pathway in the myocardium of mice after myocardial infarction.
Phytotherapy research : PTR
Tian CX,Li MY,Shuai XX,Jiang F,Dong YL,Gui Y,Zhang ZL,Qin RJ,Kang ZY,Lin L,Sarapultsev A,Wu B,Luo SS,Hu DS
Sun Jan 01 00:00:00 EST 2023
Primary intranasal perivascular wall tumors in 2 cats.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc
Godizzi F,Avallone G,Ghisleni G,Dell'Aere S,Zamboni C,Valenti P,Roccabianca P
Sun Jan 01 00:00:00 EST 2023
Targeting the mitochondrial calcium uniporter inhibits cancer progression and alleviates cisplatin resistance in esophageal squamous cell carcinoma.
International journal of oncology
Miao Y,Wang X,Lai Y,Huang Y,Yin H,Meng X,Liu H,Hou R,Lin W,Zhang X,Zhang X,Chai BC,Zhang F,Guo L,Yang S
Sat Jul 01 00:00:00 EDT 2023
Perlecan Improves Blood Spinal Cord Barrier Repair Through the Integrin β1/ROCK/MLC Pathway After Spinal Cord Injury.
Molecular neurobiology
Xie C,Wang Y,Wang J,Xu Y,Liu H,Guo J,Zhu L
Sun Jan 01 00:00:00 EST 2023
KIF15 knockdown inhibits the development of endometrial cancer by suppressing epithelial-mesenchymal transition and stemness through Wnt/β-catenin signaling.
Environmental toxicology
Huang J,Sun X,Diao G,Li R,Guo J,Han J
Tue Aug 01 00:00:00 EDT 2023
Glycyrrhizic acid alters the hyperoxidative stress-induced differentiation commitment of MSCs by activating the Wnt/β-catenin pathway to prevent SONFH.
Food & function
Xu H,Fang L,Zeng Q,Chen J,Ling H,Xia H,Ge Q,Wu C,Zou K,Wang X,Wang P,Yuan W,Dong R,Hu S,Xiao L,He B,Tong P,Jin H
Mon Jan 23 00:00:00 EST 2023
Glycyrrhizic acid alters the hyperoxidative stress-induced differentiation commitment of MSCs by activating the Wnt/β-catenin pathway to prevent SONFH.
Food & function
Xu H,Fang L,Zeng Q,Chen J,Ling H,Xia H,Ge Q,Wu C,Zou K,Wang X,Wang P,Yuan W,Dong R,Hu S,Xiao L,He B,Tong P,Jin H
Mon Jan 23 00:00:00 EST 2023
Glycyrrhizic acid alters the hyperoxidative stress-induced differentiation commitment of MSCs by activating the Wnt/β-catenin pathway to prevent SONFH.
Food & function
Xu H,Fang L,Zeng Q,Chen J,Ling H,Xia H,Ge Q,Wu C,Zou K,Wang X,Wang P,Yuan W,Dong R,Hu S,Xiao L,He B,Tong P,Jin H
Mon Jan 23 00:00:00 EST 2023
Glycyrrhizic acid alters the hyperoxidative stress-induced differentiation commitment of MSCs by activating the Wnt/β-catenin pathway to prevent SONFH.
Food & function
Xu H,Fang L,Zeng Q,Chen J,Ling H,Xia H,Ge Q,Wu C,Zou K,Wang X,Wang P,Yuan W,Dong R,Hu S,Xiao L,He B,Tong P,Jin H
Mon Jan 23 00:00:00 EST 2023
Screening of differentially methylated genes in skeletal fluorosis of rats with different types and involvement of aberrant methylation of Cthrc1.
Environmental pollution (Barking, Essex : 1987)
Ding H,Yin C,Yang M,Zhou R,Wang X,Pan X
Fri Sep 01 00:00:00 EDT 2023
Salinomycin sodium exerts anti diffuse large B-cell lymphoma activity through inhibition of LRP6-mediated Wnt/β-catenin and mTORC1 signaling.
Leukemia & lymphoma
Li L,Zeng P,Yu L,Yang J,Man J,Zhou L,Zhao L
Thu Jun 01 00:00:00 EDT 2023
XAV-939 inhibits epithelial-mesenchymal transformation in pulmonary fibrosis induced by crystalline silica via the Wnt signaling pathway.
Environmental toxicology
Lv Z,Xu H,Si X,Xu S,Li X,Li N,Zhou Q,Chang M,Yao S,Li H
Wed Feb 01 00:00:00 EST 2023
Systematic assessment of hexavalent chromium-induced damage to male fertility and the preventive role of melatonin: a longitudinal study from the translational point of view.
Molecular human reproduction
Li T,Lv Y,Wu Z,Guo M,Liu R,Zeng W,Zheng Y
Fri Jun 30 00:00:00 EDT 2023
9-Butyl-Harmol Exerts Antiviral Activity against Newcastle Disease Virus through Targeting GSK-3β and HSP90β.
Journal of virology
Wang C,Wang T,Hu R,Duan L,Hou Q,Han Y,Dai J,Wang W,Ren S,Liu H,Wang X,Xiao S,Li N,Wang J,Yang Z
Thu Mar 30 00:00:00 EDT 2023
9-Butyl-Harmol Exerts Antiviral Activity against Newcastle Disease Virus through Targeting GSK-3β and HSP90β.
Journal of virology
Wang C,Wang T,Hu R,Duan L,Hou Q,Han Y,Dai J,Wang W,Ren S,Liu H,Wang X,Xiao S,Li N,Wang J,Yang Z
Thu Mar 30 00:00:00 EDT 2023
Baicalin Attenuates Continuous Activation of β-Catenin Induced by Lipopolysaccharide (LPS) and Depression Complicated by Infertility in Male Rats.
Evidence-based complementary and alternative medicine : eCAM
Fan R,Dai F,Zhang C,Zhou L,Qiu X,Li Y
Sat Jul 16 00:00:00 EDT 2022
Up-regulation of caveolin 1 mediated by chitosan activates Wnt/ β-catenin pathway in chronic refractory wound diabetic rat model.
Bioengineered
Gao J,Zhang J,Xia L,Liang X,Ding W,Song M,Li L,Jia Z
Sat Jan 01 00:00:00 EST 2022
Enhanced effect of combining bone marrow mesenchymal stem cells (BMMSCs) and pulsed electromagnetic fields (PEMF) to promote recovery after spinal cord injury in mice.
MedComm
Huang L,Sun X,Wang L,Pei G,Wang Y,Zhang Q,Liang Z,Wang D,Fu C,He C,Wei Q
Thu Sep 01 00:00:00 EDT 2022
Enhanced effect of combining bone marrow mesenchymal stem cells (BMMSCs) and pulsed electromagnetic fields (PEMF) to promote recovery after spinal cord injury in mice.
MedComm
Huang L,Sun X,Wang L,Pei G,Wang Y,Zhang Q,Liang Z,Wang D,Fu C,He C,Wei Q
Thu Sep 01 00:00:00 EDT 2022
SKI knockdown suppresses apoptosis and extracellular matrix degradation of nucleus pulposus cells via inhibition of the Wnt/β-catenin pathway and ameliorates disc degeneration.
Apoptosis : an international journal on programmed cell death
Wu ZL,Chen YJ,Zhang GZ,Xie QQ,Wang KP,Yang X,Liu TC,Wang ZQ,Zhao GH,Zhang HH
Tue Feb 01 00:00:00 EST 2022
Overall Survival Signature of 5-Methylcytosine Regulators Related Long Non-Coding RNA in Hepatocellular Carcinoma.
Frontiers in oncology
Pan Q,Yi C,Zhang Y
Sat Jul 16 00:00:00 EDT 2022
MARCH1 promotes the growth and maintaining of stem cell-like characteristics of gastric cancer cells by activating the Wnt/β-catenin signaling pathway.
Tissue & cell
Liang W,Zhang T,Huo J,Yang J
Sat Oct 01 00:00:00 EDT 2022
Integrative network analysis of circular RNAs reveals regulatory mechanisms for hepatic specification of human iPSC-derived endoderm.
Stem cell research & therapy
Bai F,Duan J,Yang D,Lai X,Zhu X,He X,Hu A
Thu Sep 08 00:00:00 EDT 2022
FAT4 activation inhibits epithelial-mesenchymal transition (EMT) by promoting autophagy in H2228/Cer cells.
Medical oncology (Northwood, London, England)
Yang Y,Li Y,Yang Q,Liu Z,Chang X,Yang H,Liu J,Li Z,Zuo D
Wed Dec 28 00:00:00 EST 2022
TNFAIP6 Promotes Gastric Carcinoma Cell Invasion via Upregulating PTX3 and Activating the Wnt/β-Catenin Signaling Pathway.
Contrast media & molecular imaging
Cui H,Zhang L,Chen B,Zhang F,Xu H,Ma G,Cao L,Wang M
Thu Jul 21 00:00:00 EDT 2022
B-cell lymphoma-3 controls mesenchymal stem cell commitment and senescence during skeletal aging.
Clinical and translational medicine
Wang F,Guo J,Wang S,Wang Y,Chen J,Hu Y,Zhang H,Xu K,Wei Y,Cao L,Chen X,Jing Y,Su J
Fri Jul 01 00:00:00 EDT 2022
EPA and DHA differentially coordinate the crosstalk between host and gut microbiota and block DSS-induced colitis in mice by a reinforced colonic mucus barrier.
Food & function
Fang J,Zhang Z,Cheng Y,Yang H,Zhang H,Xue Z,Lu S,Dong Y,Song C,Zhang X,Zhou Y
Wed Apr 20 00:00:00 EDT 2022
Quantum dots bind nanosheet to promote nanomaterial stability and resist endotoxin-induced fibrosis and PM2.5-induced pneumonia.
Ecotoxicology and environmental safety
Zhou Q,Li D,Zhang S,Wang S,Hu X
Fri Apr 01 00:00:00 EDT 2022
Ginsenoside Rc Promotes Bone Formation in Ovariectomy-Induced Osteoporosis In Vivo and Osteogenic Differentiation In Vitro.
International journal of molecular sciences
Yang N,Zhang X,Li L,Xu T,Li M,Zhao Q,Yu J,Wang J,Liu Z
Tue May 31 00:00:00 EDT 2022
Expression of Insulin-Like Growth Factor Type 1 Receptor Is Linked to Inflammation in Adamantinomatous Craniopharyngioma.
Neuroendocrinology
Yang L,Li K,Li W,Wang C,Liu Y,Zhang H,Pan J,Qi S,Peng J
Tue Aug 23 00:00:00 EDT 2022
MICALL2 as a substrate of ubiquitinase TRIM21 regulates tumorigenesis of colorectal cancer.
Cell communication and signaling : CCS
Wen P,Wang H,Li Y,Sui X,Hou Z,Guo X,Xue W,Liu D,Wang Y,Gao J
Fri Oct 28 00:00:00 EDT 2022
Histone methyltransferase KMT2D cooperates with MEF2A to promote the stem-like properties of oral squamous cell carcinoma.
Cell & bioscience
Wang X,Li R,Wu L,Chen Y,Liu S,Zhao H,Wang Y,Wang L,Shao Z
Wed Apr 27 00:00:00 EDT 2022
Histone methyltransferase KMT2D cooperates with MEF2A to promote the stem-like properties of oral squamous cell carcinoma.
Cell & bioscience
Wang X,Li R,Wu L,Chen Y,Liu S,Zhao H,Wang Y,Wang L,Shao Z
Wed Apr 27 00:00:00 EDT 2022
Histone methyltransferase KMT2D cooperates with MEF2A to promote the stem-like properties of oral squamous cell carcinoma.
Cell & bioscience
Wang X,Li R,Wu L,Chen Y,Liu S,Zhao H,Wang Y,Wang L,Shao Z
Wed Apr 27 00:00:00 EDT 2022
Overexpression of ERCC6L correlates with poor prognosis and confers malignant phenotypes of lung adenocarcinoma.
Oncology reports
Huang X,Jiang L,Lu S,Yuan M,Lin H,Li B,Wen Z,Zhong Y
Fri Jul 01 00:00:00 EDT 2022
VCP interaction with HMGB1 promotes hepatocellular carcinoma progression by activating the PI3K/AKT/mTOR pathway.
Journal of translational medicine
Pu Z,Duda DG,Zhu Y,Pei S,Wang X,Huang Y,Yi P,Huang Z,Peng F,Hu X,Fan X
Fri May 13 00:00:00 EDT 2022
Substrate stiffness regulates differentiation of induced pluripotent stem cells into heart valve endothelial cells.
Acta biomaterialia
Cao H,Zhou Q,Liu C,Zhang Y,Xie M,Qiao W,Dong N
Fri Apr 15 00:00:00 EDT 2022
Heterozygote loss-of-function variants in the LRP5 gene cause familial exudative vitreoretinopathy.
Clinical & experimental ophthalmology
Zhao R,Wang S,Zhao P,Dai E,Zhang X,Peng L,He Y,Yang M,Li S,Yang Z
Sun May 01 00:00:00 EDT 2022
Forkhead Box S1 mediates epithelial-mesenchymal transition through the Wnt/β-catenin signaling pathway to regulate colorectal cancer progression.
Journal of translational medicine
Zhang L,Ren CF,Yang Z,Gong LB,Wang C,Feng M,Guan WX
Thu Jul 21 00:00:00 EDT 2022
N-glycosylation of CD82 at Asn157 is required for suppressing migration and invasion by reversing EMT via Wnt/β-catenin pathway in colon cancer.
Biochemical and biophysical research communications
Zhu L,Chen Y,Du H,Cong Y,Yan W,Ma K,Huang X
Sat Nov 12 00:00:00 EST 2022
MicroRNA-1246 suppresses the metastasis of breast cancer cells by targeting the DYRK1A/PGRN axis to prevent the epithelial-mesenchymal transition.
Molecular biology reports
Wang P,Chen W,Zhang Y,Zhong Q,Li Z,Wang Y
Fri Apr 01 00:00:00 EDT 2022
Protective effect of 3-n-butylphthalide against intrastriatal injection of malonic acid-induced neurotoxicity and biochemical alteration in rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Yuan C,Zheng L,Zhao Y
Tue Nov 01 00:00:00 EDT 2022
Fucoxanthin induces apoptosis and reverses epithelial-mesenchymal transition via inhibiting Wnt/β-catenin pathway in lung adenocarcinoma.
Discover. Oncology
Luan H,Yan L,Zhao Y,Ding X,Cao L
Mon Oct 03 00:00:00 EDT 2022
Wnt/β-Catenin Signaling Inhibits Osteogenic Differentiation in Human Periodontal Ligament Fibroblasts.
Biomimetics (Basel, Switzerland)
Iizumi R,Honda M
Sat Dec 03 00:00:00 EST 2022
Wnt/β-Catenin Signaling Inhibits Osteogenic Differentiation in Human Periodontal Ligament Fibroblasts.
Biomimetics (Basel, Switzerland)
Iizumi R,Honda M
Sat Dec 03 00:00:00 EST 2022
Exploratory study of sea buckthorn enhancing QiangGuYin efficacy by inhibiting CKIP-1 and Notum activating the Wnt/β-catenin signaling pathway and analysis of active ingredients by molecular docking.
Frontiers in pharmacology
Yuan YF,Wang S,Zhou H,Tang BB,Liu Y,Huang H,He CJ,Chen TP,Fang MH,Liang BC,Mao YD,Qie FQ,Liu K,Shi XL
Sat Oct 29 00:00:00 EDT 2022
LHX2 facilitates the progression of nasopharyngeal carcinoma via activation of the FGF1/FGFR axis.
British journal of cancer
Xie T,Du K,Liu W,Liu C,Wang B,Tian Y,Li R,Huang X,Lin J,Jian H,Zhang J,Yuan Y
Sat Oct 01 00:00:00 EDT 2022
HDAC9 Contributes to Serous Ovarian Cancer Progression through Regulating Epithelial-Mesenchymal Transition.
Biomedicines
Xu L,Wang J,Liu B,Fu J,Zhao Y,Yu S,Shen L,Yan X,Su J
Thu Feb 03 00:00:00 EST 2022
HDAC9 Contributes to Serous Ovarian Cancer Progression through Regulating Epithelial-Mesenchymal Transition.
Biomedicines
Xu L,Wang J,Liu B,Fu J,Zhao Y,Yu S,Shen L,Yan X,Su J
Thu Feb 03 00:00:00 EST 2022
Natural shikonin and acetyl-shikonin improve intestinal microbial and protein composition to alleviate colitis-associated colorectal cancer.
International immunopharmacology
Lin H,Ma X,Yang X,Chen Q,Wen Z,Yang M,Fu J,Yin T,Lu G,Qi J,Han H,Yang Y
Sat Oct 01 00:00:00 EDT 2022
Exosome-transmitted S100A4 induces immunosuppression and non-small cell lung cancer development by activating STAT3.
Clinical and experimental immunology
Wu X,Zhang H,Jiang G,Peng M,Li C,Lu J,Jiang S,Yang X,Jiang Y
Sat Dec 31 00:00:00 EST 2022
Novel long non-coding RNA CYB561-5 promotes aerobic glycolysis and tumorigenesis by interacting with basigin in non-small cell lung cancer.
Journal of cellular and molecular medicine
Li L,Li Z,Qu J,Wei X,Suo F,Xu J,Liu X,Chen C,Zheng S
Tue Mar 01 00:00:00 EST 2022
Selenium Deficiency Leads to Changes in Renal Fibrosis Marker Proteins and Wnt/β-Catenin Signaling Pathway Components.
Biological trace element research
Lin T,Tao J,Chen Y,Zhang Y,Li F,Zhang Y,Han X,Zhao Z,Liu G,Li H
Tue Mar 01 00:00:00 EST 2022
miR-1 Regulates Differentiation and Proliferation of Goat Hair Follicle Stem Cells by Targeting IGF1R and LEF1 Genes.
DNA and cell biology
Yan H,Jin M,Li Y,Gao Y,Ding Q,Wang X,Zeng W,Chen Y
Tue Feb 01 00:00:00 EST 2022
Astragaloside IV Alleviates Intestinal Barrier Dysfunction via the AKT-GSK3β-β-Catenin Pathway in Peritoneal Dialysis.
Frontiers in pharmacology
He J,Wang M,Yang L,Xin H,Bian F,Jiang G,Zhang X
Sat Jul 16 00:00:00 EDT 2022
Astragaloside IV Alleviates Intestinal Barrier Dysfunction via the AKT-GSK3β-β-Catenin Pathway in Peritoneal Dialysis.
Frontiers in pharmacology
He J,Wang M,Yang L,Xin H,Bian F,Jiang G,Zhang X
Sat Jul 16 00:00:00 EDT 2022
Astragaloside IV Alleviates Intestinal Barrier Dysfunction via the AKT-GSK3β-β-Catenin Pathway in Peritoneal Dialysis.
Frontiers in pharmacology
He J,Wang M,Yang L,Xin H,Bian F,Jiang G,Zhang X
Sat Jul 16 00:00:00 EDT 2022
Astragaloside IV Alleviates Intestinal Barrier Dysfunction via the AKT-GSK3β-β-Catenin Pathway in Peritoneal Dialysis.
Frontiers in pharmacology
He J,Wang M,Yang L,Xin H,Bian F,Jiang G,Zhang X
Sat Jul 16 00:00:00 EDT 2022
Astragaloside IV Alleviates Intestinal Barrier Dysfunction via the AKT-GSK3β-β-Catenin Pathway in Peritoneal Dialysis.
Frontiers in pharmacology
He J,Wang M,Yang L,Xin H,Bian F,Jiang G,Zhang X
Sat Jul 16 00:00:00 EDT 2022
Astragaloside IV Alleviates Intestinal Barrier Dysfunction via the AKT-GSK3β-β-Catenin Pathway in Peritoneal Dialysis.
Frontiers in pharmacology
He J,Wang M,Yang L,Xin H,Bian F,Jiang G,Zhang X
Sat Jul 16 00:00:00 EDT 2022
The E3 Ubiquitin Ligase TRIM11 Facilitates Gastric Cancer Progression by Activating the Wnt/β-Catenin Pathway via Destabilizing Axin1 Protein.
Journal of oncology
Zhou L,Wang H,Zhong M,Fang Z,Le Y,Nie F,Zhou J,Xiong J,Xiang X,Fang Z
Mon May 02 00:00:00 EDT 2022
The combination of decitabine and aspirin inhibits tumor growth and metastasis in non-small cell lung cancer.
The Journal of international medical research
Xu M,Song B,Yang X,Li N
Fri Jul 01 00:00:00 EDT 2022
Insulin and liraglutide attenuate brain pathology in diabetic mice by enhancing the Wnt/β-catenin signaling pathway.
Experimental and therapeutic medicine
Zhao Y,Yu J,Ping F,Xu L,Li W,Zhang H,Li Y
Fri Jul 01 00:00:00 EDT 2022
Combination of curaxin and tyrosine kinase inhibitors display enhanced killing of primitive Chronic Myeloid Leukaemia cells.
PloS one
Pearson S,Whetton AD,Pierce A
Thu Apr 14 00:00:00 EDT 2022
Epigenetic silencing of JAM3 promotes esophageal cancer development by activating Wnt signaling.
Clinical epigenetics
Yang W,Guo C,Herman JG,Zhu C,Lv H,Su X,Zhang L,Zhang M,Guo M
Fri Dec 02 00:00:00 EST 2022
Epigenetic silencing of JAM3 promotes esophageal cancer development by activating Wnt signaling.
Clinical epigenetics
Yang W,Guo C,Herman JG,Zhu C,Lv H,Su X,Zhang L,Zhang M,Guo M
Fri Dec 02 00:00:00 EST 2022
Epigenetic silencing of JAM3 promotes esophageal cancer development by activating Wnt signaling.
Clinical epigenetics
Yang W,Guo C,Herman JG,Zhu C,Lv H,Su X,Zhang L,Zhang M,Guo M
Fri Dec 02 00:00:00 EST 2022
Epigenetic silencing of JAM3 promotes esophageal cancer development by activating Wnt signaling.
Clinical epigenetics
Yang W,Guo C,Herman JG,Zhu C,Lv H,Su X,Zhang L,Zhang M,Guo M
Fri Dec 02 00:00:00 EST 2022
Chromogranin A-derived peptide CGA47-66 protects against septic brain injury by reducing blood-brain barrier damage through the PI3K/AKT pathway.
Biochemical and biophysical research communications
Wang F,Zeng Y,Liu X,Cao J,Kang S,Zhou W,Chen X,Liu J,Zhang D
Sat May 21 00:00:00 EDT 2022
SP2509, a Selective Inhibitor of LSD1, Suppresses Retinoblastoma Growth by Downregulating β-catenin Signaling.
Investigative ophthalmology & visual science
Jiang A,Wu W,Xu C,Mao L,Ao S,Guo H,Sun X,Tao J,Sang Y,Huang G
Wed Mar 02 00:00:00 EST 2022
miR-145-5p targets MMP2 to protect brain injury in hypertensive intracerebral hemorrhage via inactivation of the Wnt/β-catenin signaling pathway.
Annals of translational medicine
Xiao W,Jiang Z,Wan W,Pan W,Xu J
Sun May 01 00:00:00 EDT 2022
FAT1 downregulation enhances stemness and cisplatin resistance in esophageal squamous cell carcinoma.
Molecular and cellular biochemistry
Zhai Y,Shan C,Zhang H,Kong P,Zhang L,Wang Y,Hu X,Cheng X
Thu Dec 01 00:00:00 EST 2022
FAT1 downregulation enhances stemness and cisplatin resistance in esophageal squamous cell carcinoma.
Molecular and cellular biochemistry
Zhai Y,Shan C,Zhang H,Kong P,Zhang L,Wang Y,Hu X,Cheng X
Thu Dec 01 00:00:00 EST 2022
FAT1 downregulation enhances stemness and cisplatin resistance in esophageal squamous cell carcinoma.
Molecular and cellular biochemistry
Zhai Y,Shan C,Zhang H,Kong P,Zhang L,Wang Y,Hu X,Cheng X
Thu Dec 01 00:00:00 EST 2022
FAT1 downregulation enhances stemness and cisplatin resistance in esophageal squamous cell carcinoma.
Molecular and cellular biochemistry
Zhai Y,Shan C,Zhang H,Kong P,Zhang L,Wang Y,Hu X,Cheng X
Thu Dec 01 00:00:00 EST 2022
FAT1 downregulation enhances stemness and cisplatin resistance in esophageal squamous cell carcinoma.
Molecular and cellular biochemistry
Zhai Y,Shan C,Zhang H,Kong P,Zhang L,Wang Y,Hu X,Cheng X
Thu Dec 01 00:00:00 EST 2022
FAT1 downregulation enhances stemness and cisplatin resistance in esophageal squamous cell carcinoma.
Molecular and cellular biochemistry
Zhai Y,Shan C,Zhang H,Kong P,Zhang L,Wang Y,Hu X,Cheng X
Thu Dec 01 00:00:00 EST 2022
Doxorubicin promotes breast cancer cell migration and invasion via DCAF13.
FEBS open bio
Sun Z,Zhou D,Yang J,Zhang D
Sat Jan 01 00:00:00 EST 2022
Doxorubicin promotes breast cancer cell migration and invasion via DCAF13.
FEBS open bio
Sun Z,Zhou D,Yang J,Zhang D
Sat Jan 01 00:00:00 EST 2022
G6PD promotes cell proliferation and dexamethasone resistance in multiple myeloma via increasing anti-oxidant production and activating Wnt/β-catenin pathway.
Experimental hematology & oncology
Li R,Ke M,Qi M,Han Z,Cao Y,Deng Z,Qian J,Yang Y,Gu C
Fri Oct 21 00:00:00 EDT 2022
LncRNA FAM83H-AS1 promotes the malignant progression of pancreatic ductal adenocarcinoma by stabilizing FAM83H mRNA to protect β-catenin from degradation.
Journal of experimental & clinical cancer research : CR
Zhou M,Pan S,Qin T,Zhao C,Yin T,Gao Y,Liu Y,Zhang Z,Shi Y,Bai Y,Gong J,Guo X,Wang M,Qin R
Thu Sep 29 00:00:00 EDT 2022
Knockdown of CDC20 promotes adipogenesis of bone marrow-derived stem cells by modulating β-catenin.
Stem cell research & therapy
Du Y,Liu Y,Zhou Y,Zhang P
Fri Sep 02 00:00:00 EDT 2022
MicroRNA-20a promotes non-small cell lung cancer proliferation by upregulating PD-L1 by targeting PTEN.
Oncology letters
Gong J,Shen Y,Jiang F,Wang Y,Chu L,Sun J,Shen P,Chen M
Sun May 01 00:00:00 EDT 2022
Fatty acid-binding protein 5 aggravates pulmonary artery fibrosis in pulmonary hypertension secondary to left heart disease via activating wnt/β-catenin pathway.
Journal of advanced research
Lei Q,Yu Z,Li H,Cheng J,Wang Y
Thu Sep 01 00:00:00 EDT 2022
Thymopentin-Mediated Inhibition of Cancer Stem Cell Stemness Enhances the Cytotoxic Effect of Oxaliplatin on Colon Cancer Cells.
Frontiers in pharmacology
Yu PC,Liu D,Han ZX,Liang F,Hao CY,Lei YT,Guo CR,Wang WH,Li XH,Yang XN,Li CZ,Yu Y,Fan YZ
Sat Mar 05 00:00:00 EST 2022
Luteolin alleviates ischemia/reperfusion injury-induced no-reflow by regulating Wnt/β-catenin signaling in rats.
Microvascular research
Qin X,Qin H,Li Z,Xue S,Huang B,Liu X,Wang D
Sat Jan 01 00:00:00 EST 2022
Phase II Study of Gemcitabine, Peg-Asparaginase, Dexamethasone and Methotrexate Regimen for Newly Diagnosed Extranodal Natural Killer/T-Cell Lymphoma, Nasal Type: Final Analysis With Long-Term Follow-Up and Rational Research for the Combination.
Frontiers in oncology
Wang Y,Wang CQ,Sun P,Liu PP,Yang H,Wang HY,Rao HL,Li S,Jiang WQ,Huang JJ,Li ZM
Sun May 01 00:00:00 EDT 2022
ALG-bFGF Hydrogel Inhibiting Autophagy Contributes to Protection of Blood-Spinal Cord Barrier Integrity via PI3K/Akt/FOXO1/KLF4 Pathway After SCI.
Frontiers in pharmacology
Zhang R,Xie L,Wu F,Xu J,Lu L,Cao L,Li L,Meng W,Zhang H,Shao C,Li X,Chen D
Tue Mar 29 00:00:00 EDT 2022
ANGPTL8 promotes adipogenic differentiation of mesenchymal stem cells: potential role in ectopic lipid deposition.
Frontiers in endocrinology
Tang J,Ma S,Gao Y,Zeng F,Feng Y,Guo C,Hu L,Yang L,Chen Y,Zhang Q,Yuan Y,Guo X
Tue Aug 30 00:00:00 EDT 2022
ANGPTL8 promotes adipogenic differentiation of mesenchymal stem cells: potential role in ectopic lipid deposition.
Frontiers in endocrinology
Tang J,Ma S,Gao Y,Zeng F,Feng Y,Guo C,Hu L,Yang L,Chen Y,Zhang Q,Yuan Y,Guo X
Tue Aug 30 00:00:00 EDT 2022
Fufang Xueshuantong Improves Diabetic Cardiomyopathy by Regulating the Wnt/β-Catenin Pathway.
International journal of endocrinology
Peng M,Liu H,Ji Q,Ma P,Niu Y,Ning S,Sun H,Pang X,Yang Y,Zhang Y,Han J,Hao G
Sat Oct 15 00:00:00 EDT 2022
The ameliorative effect of melatonin on LPS-induced Sertoli cells inflammatory and tight junctions damage via suppression of the TLR4/MyD88/NF-κB signaling pathway in newborn calf.
Theriogenology
Feng R,Adeniran SO,Huang F,Li Y,Ma M,Zheng P,Zhang G
Tue Feb 01 00:00:00 EST 2022
GALNT1 Enhances Malignant Phenotype of Gastric Cancer via Modulating CD44 Glycosylation to Activate the Wnt/β-catenin Signaling Pathway.
International journal of biological sciences
Zhang J,Wang H,Wu J,Yuan C,Chen S,Liu S,Huo M,Zhang C,He Y
Tue Nov 29 00:00:00 EST 2022
GALNT1 Enhances Malignant Phenotype of Gastric Cancer via Modulating CD44 Glycosylation to Activate the Wnt/β-catenin Signaling Pathway.
International journal of biological sciences
Zhang J,Wang H,Wu J,Yuan C,Chen S,Liu S,Huo M,Zhang C,He Y
Tue Nov 29 00:00:00 EST 2022
Blue Light Alters the Composition of the Jejunal Microbiota and Promotes the Development of the Small Intestine by Reducing Oxidative Stress.
Antioxidants (Basel, Switzerland)
Zhang Y,Wang Z,Dong Y,Cao J,Chen Y
Sat Jan 29 00:00:00 EST 2022
Circular RNA circSHPRH inhibits the malignant behaviors of bladder cancer by regulating the miR-942/BARX2 pathway.
Aging
Zuo L,Zhu Y,Han J,Liu H
Thu Feb 24 00:00:00 EST 2022
A novel Granzymes and miRNA nanocapsules co-delivery system for tumor suppression.
Biomedical physics & engineering express
Shi Z,Zhao M,Lin T,Chen J,Qian X
Fri Nov 11 00:00:00 EST 2022
A novel Granzymes and miRNA nanocapsules co-delivery system for tumor suppression.
Biomedical physics & engineering express
Shi Z,Zhao M,Lin T,Chen J,Qian X
Fri Nov 11 00:00:00 EST 2022
A novel Granzymes and miRNA nanocapsules co-delivery system for tumor suppression.
Biomedical physics & engineering express
Shi Z,Zhao M,Lin T,Chen J,Qian X
Fri Nov 11 00:00:00 EST 2022
A novel Granzymes and miRNA nanocapsules co-delivery system for tumor suppression.
Biomedical physics & engineering express
Shi Z,Zhao M,Lin T,Chen J,Qian X
Fri Nov 11 00:00:00 EST 2022
MICAL1 inhibits colorectal cancer cell migration and proliferation by regulating the EGR1/β-catenin signaling pathway.
Biochemical pharmacology
Gu H,Li Y,Cui X,Cao H,Hou Z,Ti Y,Liu D,Gao J,Wang Y,Wen P
Sat Jan 01 00:00:00 EST 2022
MICAL1 inhibits colorectal cancer cell migration and proliferation by regulating the EGR1/β-catenin signaling pathway.
Biochemical pharmacology
Gu H,Li Y,Cui X,Cao H,Hou Z,Ti Y,Liu D,Gao J,Wang Y,Wen P
Sat Jan 01 00:00:00 EST 2022
PKC-ζ Aggravates Doxorubicin-Induced Cardiotoxicity by Inhibiting Wnt/β-Catenin Signaling.
Frontiers in pharmacology
Cao YJ,Li JY,Wang PX,Lin ZR,Yu WJ,Zhang JG,Lu J,Liu PQ
Fri Mar 04 00:00:00 EST 2022
N-myc downstream regulated gene 1 suppresses osteoblast differentiation through inactivating Wnt/β-catenin signaling.
Stem cell research & therapy
Shi X,Cen Y,Shan L,Tian L,Zhu E,Yuan H,Li X,Liu Y,Wang B
Fri Feb 04 00:00:00 EST 2022
STARD3NL inhibits the osteogenic differentiation by inactivating the Wnt/β-catenin pathway via binding to Annexin A2 in osteoporosis.
Journal of cellular and molecular medicine
Xu Y,Bao X,Chen X,Wu P,Chen S,Zhang B,Ma J,Xu G,Ma D
Tue Mar 01 00:00:00 EST 2022
Nc886 promotes renal cancer cell drug-resistance by enhancing EMT through Rock2 phosphorylation-mediated β-catenin nuclear translocation.
Cell cycle (Georgetown, Tex.)
Gao W,Zhang S,Guorong L,Liu Q,Zhu A,Gui F,Zou Y,Wu Y,Luo Y,Hong Z
Tue Feb 01 00:00:00 EST 2022
DDX3X alleviates doxorubicin-induced cardiotoxicity by regulating Wnt/β-catenin signaling pathway in an in vitro model.
Journal of biochemical and molecular toxicology
Feng D,Li J,Guo L,Liu J,Wang S,Ma X,Song Y,Liu J,Hao E
Mon Aug 01 00:00:00 EDT 2022
UBE2S as a novel ubiquitinated regulator of p16 and β-catenin to promote bone metastasis of prostate cancer.
International journal of biological sciences
Peng S,Chen X,Huang C,Yang C,Situ M,Zhou Q,Ling Y,Huang H,Huang M,Zhang Y,Cheng L,Zhang Q,Guo Z,Lai Y,Huang J
Thu Jun 02 00:00:00 EDT 2022
Cholecystectomy promotes colon carcinogenesis by activating the Wnt signaling pathway by increasing the deoxycholic acid level.
Cell communication and signaling : CCS
Yao Y,Li X,Xu B,Luo L,Guo Q,Wang X,Sun L,Zhang Z,Li P
Wed May 25 00:00:00 EDT 2022
Pan-cancer analysis, cell and animal experiments revealing TEAD4 as a tumor promoter in ccRCC.
Life sciences
Li F,Feng Y,Jiang Q,Zhang J,Wu F,Li Q,Jing X,Wang X,Huang C
Tue Mar 15 00:00:00 EDT 2022
CSTF2 Acts as a Prognostic Marker Correlated with Immune Infiltration in Hepatocellular Carcinoma.
Cancer management and research
Zhang W,Wan Y,Zhang Y,Liu Q,Zhu X
Tue Sep 20 00:00:00 EDT 2022
Sertoli cell and spermatogonial development in pigs.
Journal of animal science and biotechnology
Zheng Y,Gao Q,Li T,Liu R,Cheng Z,Guo M,Xiao J,Wu D,Zeng W
Mon Apr 11 00:00:00 EDT 2022
Inhibition of GABAAR or Application of Lactobacillus casei Zhang Alleviates Ulcerative Colitis in Mice: GABAAR as a Potential Target for Intestinal Epithelial Renewal and Repair.
International journal of molecular sciences
Wang Q,Deng Z,Lan J,Li D,Fan K,Chang J,Ma Y
Fri Sep 23 00:00:00 EDT 2022
Blockade of Nuclear β-Catenin Signaling via Direct Targeting of RanBP3 with NU2058 Induces Cell Senescence to Suppress Colorectal Tumorigenesis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zheng CC,Liao L,Liu YP,Yang YM,He Y,Zhang GG,Li SJ,Liu T,Xu WW,Li B
Thu Dec 01 00:00:00 EST 2022
Blockade of Nuclear β-Catenin Signaling via Direct Targeting of RanBP3 with NU2058 Induces Cell Senescence to Suppress Colorectal Tumorigenesis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zheng CC,Liao L,Liu YP,Yang YM,He Y,Zhang GG,Li SJ,Liu T,Xu WW,Li B
Thu Dec 01 00:00:00 EST 2022
Blockade of Nuclear β-Catenin Signaling via Direct Targeting of RanBP3 with NU2058 Induces Cell Senescence to Suppress Colorectal Tumorigenesis.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Zheng CC,Liao L,Liu YP,Yang YM,He Y,Zhang GG,Li SJ,Liu T,Xu WW,Li B
Thu Dec 01 00:00:00 EST 2022
TRIM36 inhibits tumorigenesis through the Wnt/β-catenin pathway and promotes caspase-dependent apoptosis in hepatocellular carcinoma.
Cancer cell international
Tong Q,Yi M,Kong P,Xu L,Huang W,Niu Y,Gan X,Zhan H,Tian R,Yan D
Tue Sep 06 00:00:00 EDT 2022
TRIM36 inhibits tumorigenesis through the Wnt/β-catenin pathway and promotes caspase-dependent apoptosis in hepatocellular carcinoma.
Cancer cell international
Tong Q,Yi M,Kong P,Xu L,Huang W,Niu Y,Gan X,Zhan H,Tian R,Yan D
Tue Sep 06 00:00:00 EDT 2022
SNORA70E promotes the occurrence and development of ovarian cancer through pseudouridylation modification of RAP1B and alternative splicing of PARPBP.
Journal of cellular and molecular medicine
Chen S,Li QH,Chen X,Bao HJ,Wu W,Shen F,Lu BF,Jiang RQ,Zong ZH,Zhao Y
Sat Oct 01 00:00:00 EDT 2022
SEC14L3 plays a tumor-suppressive role in breast cancer through a Wnt/β-catenin-related way.
Experimental cell research
Zhu Q,Wan NB,Deng HW,Lu LL,Zhang Y,He X,Liu H,He Y
Mon Aug 01 00:00:00 EDT 2022
Overexpression of PTPRZ1 Regulates p120/β-Catenin Phosphorylation to Promote Carcinogenesis of Oral Submucous Fibrosis.
Journal of oncology
Ma L,Shen T,Peng H,Wu J,Wang W,Gao X
Mon May 02 00:00:00 EDT 2022
Hesperidin partly ameliorates the decabromodiphenyl ether-induced reproductive toxicity in pubertal mice.
Environmental science and pollution research international
Li S,Che S,Chen S,Ruan Z,Zhang L
Thu Dec 01 00:00:00 EST 2022
ROS/PI3K/Akt and Wnt/β-catenin signalings activate HIF-1α-induced metabolic reprogramming to impart 5-fluorouracil resistance in colorectal cancer.
Journal of experimental & clinical cancer research : CR
Dong S,Liang S,Cheng Z,Zhang X,Luo L,Li L,Zhang W,Li S,Xu Q,Zhong M,Zhu J,Zhang G,Hu S
Sat Jan 08 00:00:00 EST 2022
ROS/PI3K/Akt and Wnt/β-catenin signalings activate HIF-1α-induced metabolic reprogramming to impart 5-fluorouracil resistance in colorectal cancer.
Journal of experimental & clinical cancer research : CR
Dong S,Liang S,Cheng Z,Zhang X,Luo L,Li L,Zhang W,Li S,Xu Q,Zhong M,Zhu J,Zhang G,Hu S
Sat Jan 08 00:00:00 EST 2022
ROS/PI3K/Akt and Wnt/β-catenin signalings activate HIF-1α-induced metabolic reprogramming to impart 5-fluorouracil resistance in colorectal cancer.
Journal of experimental & clinical cancer research : CR
Dong S,Liang S,Cheng Z,Zhang X,Luo L,Li L,Zhang W,Li S,Xu Q,Zhong M,Zhu J,Zhang G,Hu S
Sat Jan 08 00:00:00 EST 2022
Alpha-Momorcharin Inhibits Proinflammatory Cytokine Expression by M1 Macrophages but Not Anti-Inflammatory Cytokine Expression by M2 Macrophages.
Journal of inflammation research
Peng K,Deng N,Meng Y,He Q,Meng H,Luo T,Wei Y,Kang Y,Zhou X,Shen F
Tue Sep 06 00:00:00 EDT 2022
A long noncoding RNA GTF2IRD2P1 suppresses cell proliferation in bladder cancer by inhibiting the Wnt/β‑catenin signaling pathway.
PeerJ
Huang Z,Gao H,Qing L,Wang B,He C,Luo N,Lu C,Fan S,Gu P,Zhao H
Wed Jan 11 00:00:00 EST 2023
The transcript ENST00000444125 of lncRNA LINC01503 promotes cancer stem cell properties of glioblastoma cells via reducing FBXW1 mediated GLI2 degradation.
Experimental cell research
Wei P,Jiang J,Xiao M,Zeng M,Liu X,Zhao B,Chen F
Tue Mar 01 00:00:00 EST 2022
Cancer-derived exosomal circ_0038138 enhances glycolysis, growth, and metastasis of gastric adenocarcinoma via the miR-198/EZH2 axis.
Translational oncology
Zheng Y,Li P,Ma J,Yang C,Dai S,Zhao C
Tue Nov 01 00:00:00 EDT 2022
Cancer-derived exosomal circ_0038138 enhances glycolysis, growth, and metastasis of gastric adenocarcinoma via the miR-198/EZH2 axis.
Translational oncology
Zheng Y,Li P,Ma J,Yang C,Dai S,Zhao C
Tue Nov 01 00:00:00 EDT 2022
Splicing factor arginine/serine-rich 8 promotes multiple myeloma malignancy and bone lesion through alternative splicing of CACYBP and exosome-based cellular communication.
Clinical and translational medicine
Zhang Y,Yu X,Sun R,Min J,Tang X,Lin Z,Xie S,Li X,Lu S,Tian Z,Gu C,Teng L,Yang Y
Tue Feb 01 00:00:00 EST 2022
Splicing factor arginine/serine-rich 8 promotes multiple myeloma malignancy and bone lesion through alternative splicing of CACYBP and exosome-based cellular communication.
Clinical and translational medicine
Zhang Y,Yu X,Sun R,Min J,Tang X,Lin Z,Xie S,Li X,Lu S,Tian Z,Gu C,Teng L,Yang Y
Tue Feb 01 00:00:00 EST 2022
METTL3-mediated m6A modification of STEAP2 mRNA inhibits papillary thyroid cancer progress by blocking the Hedgehog signaling pathway and epithelial-to-mesenchymal transition.
Cell death & disease
Zhu Y,Peng X,Zhou Q,Tan L,Zhang C,Lin S,Long M
Mon Apr 18 00:00:00 EDT 2022
Indolethylamine-N-Methyltransferase Inhibits Proliferation and Promotes Apoptosis of Human Prostate Cancer Cells: A Mechanistic Exploration.
Frontiers in cell and developmental biology
Jianfeng W,Yutao W,Jianbin B
Tue Mar 08 00:00:00 EST 2022
Overexpression of PYGO1 promotes early cardiac lineage development in human umbilical cord mesenchymal stromal/stem cells by activating the Wnt/β-catenin pathway.
Human cell
Shen J,Wu X,Zhu P,Zhuang J,Qin B,Sun F,Yuan W,Fan X,Jiang Z,Li F,Li Y,Wang Y,Zhao M
Tue Nov 01 00:00:00 EDT 2022
Long Noncoding RNA Metastasis-Associated Lung Adenocarcinoma Transcript 1 in Extracellular Vesicles Promotes Hepatic Stellate Cell Activation, Liver Fibrosis and β-Catenin Signaling Pathway.
Frontiers in physiology
Wang T,Zhang C,Meng X,Zhu B,Wang S,Yuan W,Zhang S,Xu J,Zhang C
Fri Mar 04 00:00:00 EST 2022
Long Noncoding RNA Metastasis-Associated Lung Adenocarcinoma Transcript 1 in Extracellular Vesicles Promotes Hepatic Stellate Cell Activation, Liver Fibrosis and β-Catenin Signaling Pathway.
Frontiers in physiology
Wang T,Zhang C,Meng X,Zhu B,Wang S,Yuan W,Zhang S,Xu J,Zhang C
Fri Mar 04 00:00:00 EST 2022
AR-regulated ZIC5 contributes to the aggressiveness of prostate cancer.
Cell death discovery
Tan YF,Zhang Y,Ge SY,Zhong F,Sun CY,Xia GW
Tue Sep 20 00:00:00 EDT 2022
AR-regulated ZIC5 contributes to the aggressiveness of prostate cancer.
Cell death discovery
Tan YF,Zhang Y,Ge SY,Zhong F,Sun CY,Xia GW
Tue Sep 20 00:00:00 EDT 2022
AR-regulated ZIC5 contributes to the aggressiveness of prostate cancer.
Cell death discovery
Tan YF,Zhang Y,Ge SY,Zhong F,Sun CY,Xia GW
Tue Sep 20 00:00:00 EDT 2022
Long non-coding RNA NRSN2-AS1, transcribed by SOX2, promotes progression of esophageal squamous cell carcinoma by regulating the ubiquitin-degradation of PGK1.
Clinical & experimental metastasis
Xu T,Yan Z,Lu J,Chen L,Li X,Li Y,Dong Z,Guo W
Sat Oct 01 00:00:00 EDT 2022
Loss of exosomal micro-RNA-200b-3p from hypoxia cancer-associated fibroblasts reduces sensitivity to 5-flourouracil in colorectal cancer through targeting high-mobility group box 3.
Frontiers in oncology
Yuan H,Chen B,Chai R,Gong W,Wan Z,Zheng B,Hu X,Guo Y,Gao S,Dai Q,Yu P,Tu S
Tue Oct 25 00:00:00 EDT 2022
Sclerostin Suppression Facilitates Uveal Melanoma Progression Through Activating Wnt/β-Catenin Signaling Via Binding to Membrane Receptors LRP5/LRP6.
Frontiers in oncology
Wang H,Zhao S,Liu Y,Sun F,Huang X,Wu T
Sat Jul 16 00:00:00 EDT 2022
Sclerostin Suppression Facilitates Uveal Melanoma Progression Through Activating Wnt/β-Catenin Signaling Via Binding to Membrane Receptors LRP5/LRP6.
Frontiers in oncology
Wang H,Zhao S,Liu Y,Sun F,Huang X,Wu T
Sat Jul 16 00:00:00 EDT 2022
Metastasis suppressor 1 controls osteoblast differentiation and bone homeostasis through regulating Src-Wnt/β-catenin signaling.
Cellular and molecular life sciences : CMLS
Chen M,Shan L,Gan Y,Tian L,Zhou J,Zhu E,Yuan H,Li X,Wang B
Sun Jan 30 00:00:00 EST 2022
NSUN2-mediated RNA m5C modification modulates uveal melanoma cell proliferation and migration.
Epigenetics
Luo G,Xu W,Chen X,Wang S,Wang J,Dong F,Hu DN,Reinach PS,Yan D
Mon Aug 01 00:00:00 EDT 2022
Cancer-associated fibroblasts promote the stemness and progression of renal cell carcinoma via exosomal miR-181d-5p.
Cell death discovery
Ding M,Zhao X,Chen X,Diao W,Kan Y,Cao W,Chen W,Jiang B,Qin H,Gao J,Zhuang J,Zhang Q,Guo H
Tue Nov 01 00:00:00 EDT 2022
Cancer-associated fibroblasts promote the stemness and progression of renal cell carcinoma via exosomal miR-181d-5p.
Cell death discovery
Ding M,Zhao X,Chen X,Diao W,Kan Y,Cao W,Chen W,Jiang B,Qin H,Gao J,Zhuang J,Zhang Q,Guo H
Tue Nov 01 00:00:00 EDT 2022
Cancer-associated fibroblasts promote the stemness and progression of renal cell carcinoma via exosomal miR-181d-5p.
Cell death discovery
Ding M,Zhao X,Chen X,Diao W,Kan Y,Cao W,Chen W,Jiang B,Qin H,Gao J,Zhuang J,Zhang Q,Guo H
Tue Nov 01 00:00:00 EDT 2022
Cancer-associated fibroblasts promote the stemness and progression of renal cell carcinoma via exosomal miR-181d-5p.
Cell death discovery
Ding M,Zhao X,Chen X,Diao W,Kan Y,Cao W,Chen W,Jiang B,Qin H,Gao J,Zhuang J,Zhang Q,Guo H
Tue Nov 01 00:00:00 EDT 2022
Ovarian tumorB1-mediated heat shock transcription factor 1 deubiquitination is critical for glycolysis and development of endometriosis.
iScience
Ling X,Lu J,Wang X,Liu L,Liu L,Wang Y,Sun Y,Ren C,Lu C,Yu Z
Fri Nov 18 00:00:00 EST 2022
Ovarian tumorB1-mediated heat shock transcription factor 1 deubiquitination is critical for glycolysis and development of endometriosis.
iScience
Ling X,Lu J,Wang X,Liu L,Liu L,Wang Y,Sun Y,Ren C,Lu C,Yu Z
Fri Nov 18 00:00:00 EST 2022
Anti-inflammatory potential of stevia residue extract against uric acid-associated renal injury in mice.
Journal of food biochemistry
Mehmood A,Althobaiti F,Zhao L,Usman M,Chen X,Alharthi F,Soliman MM,Shah AA,Murtaza MA,Nadeem M,Ranjha MMAN,Wang C
Sat Oct 01 00:00:00 EDT 2022
The canonical Wnt/β-catenin signaling pathway facilitates pseudorabies virus proliferation and enhances virus-induced autophagy.
Veterinary microbiology
Wang C,Hu R,Duan L,Hou Q,Yang M,Wang T,Liu H,Xiao S,Dang R,Wang J,Wang X,Zhang S,Yang Z
Thu Sep 01 00:00:00 EDT 2022
The canonical Wnt/β-catenin signaling pathway facilitates pseudorabies virus proliferation and enhances virus-induced autophagy.
Veterinary microbiology
Wang C,Hu R,Duan L,Hou Q,Yang M,Wang T,Liu H,Xiao S,Dang R,Wang J,Wang X,Zhang S,Yang Z
Thu Sep 01 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
ENKUR expression induced by chemically synthesized cinobufotalin suppresses malignant activities of hepatocellular carcinoma by modulating β-catenin/c-Jun/MYH9/USP7/c-Myc axis.
International journal of biological sciences
Hou R,Li Y,Luo X,Zhang W,Yang H,Zhang Y,Liu J,Liu S,Han S,Liu C,Huang Y,Liu Z,Li A,Fang W
Thu Apr 14 00:00:00 EDT 2022
LncRNA DACH1 protects against pulmonary fibrosis by binding to SRSF1 to suppress CTNNB1 accumulation.
Acta pharmaceutica Sinica. B
Sun J,Jin T,Niu Z,Guo J,Guo Y,Yang R,Wang Q,Gao H,Zhang Y,Li T,He W,Li Z,Ma W,Su W,Li L,Fan X,Shan H,Liang H
Thu Sep 01 00:00:00 EDT 2022
Ginsenoside Rg1 promotes remyelination and functional recovery in demyelinating disease by enhancing oligodendrocyte precursor cells-mediated myelin repair.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Liu L,Du X,Yang Q,Li M,Ran Q,Liu Q,Yang L,Sun L,Guo Y,Li Y,Chen Y,Zhu X,Li Q
Tue Nov 01 00:00:00 EDT 2022
Alpha-linolenic acid inhibits hepatocellular carcinoma cell growth through Farnesoid X receptor/β-catenin signaling pathway.
Nutrition & metabolism
Feng S,Xie X,Chen C,Zuo S,Zhao X,Li H
Tue Aug 23 00:00:00 EDT 2022
Identification of endocrine-disrupting chemicals targeting the genes and pathways of genital anomalies in males.
Ecotoxicology and environmental safety
Zhou X,Zhang X,Zhou X,Abulimiti G,Wang Y,Zhang Q,Cong R,Ji C,Luan J,Yao L,Yang J,Song N
Thu Dec 01 00:00:00 EST 2022
MFN2 knockdown promotes osteogenic differentiation of iPSC-MSCs through aerobic glycolysis mediated by the Wnt/β-catenin signaling pathway.
Stem cell research & therapy
Deng L,Yi S,Yin X,Li Y,Luan Q
Tue Apr 12 00:00:00 EDT 2022
Human CPTP promotes growth and metastasis via sphingolipid metabolite ceramide and PI4KA/AKT signaling in pancreatic cancer cells.
International journal of biological sciences
Zhang Y,Ji S,Zhang X,Lu M,Hu Y,Han Y,Shui G,Lam SM,Zou X
Mon Aug 22 00:00:00 EDT 2022
miR-141-3p affects β-catenin signaling and apoptosis by targeting Ubtd2 in rats with anorectal malformations.
Annals of the New York Academy of Sciences
Wang CY,Li SY,Xiao YX,Zhen L,Wei XG,Tang XB,Yuan ZW,Bai YZ
Thu Dec 01 00:00:00 EST 2022
Inhibition of translocator protein 18 kDa suppressed the progression of glioma via the ELAV-like RNA-binding protein 1/MAPK-activated protein kinase 3 axis.
Bioengineered
Wang J,Ren P,Zeng Z,Ma L,Li Y,Zhang H,Guo W
Tue Mar 01 00:00:00 EST 2022
Inhibition of translocator protein 18 kDa suppressed the progression of glioma via the ELAV-like RNA-binding protein 1/MAPK-activated protein kinase 3 axis.
Bioengineered
Wang J,Ren P,Zeng Z,Ma L,Li Y,Zhang H,Guo W
Tue Mar 01 00:00:00 EST 2022
METTL3-mediated m6A RNA methylation regulates dorsal lingual epithelium homeostasis.
International journal of oral science
Xiong Q,Liu C,Zheng X,Zhou X,Lei K,Zhang X,Wang Q,Lin W,Tong R,Xu R,Yuan Q
Tue May 17 00:00:00 EDT 2022
PD-L1/PD-L1 signalling promotes colorectal cancer cell migration ability through RAS/MEK/ERK.
Clinical and experimental pharmacology & physiology
Cao Y,Liang W,Fang L,Liu MK,Zuo J,Peng YL,Shan JJ,Sun RX,Zhao J,Wang J
Thu Dec 01 00:00:00 EST 2022
Microfluidic platform for studying the anti-cancer effect of ursolic acid on tumor spheroid.
Electrophoresis
Chang S,Wen J,Su Y,Ma H
Fri Jul 01 00:00:00 EDT 2022
KCTD9 inhibits the Wnt/β-catenin pathway by decreasing the level of β-catenin in colorectal cancer.
Cell death & disease
Yao H,Ren D,Wang Y,Wu L,Wu Y,Wang W,Li Q,Liu L
Fri Sep 02 00:00:00 EDT 2022
KCTD9 inhibits the Wnt/β-catenin pathway by decreasing the level of β-catenin in colorectal cancer.
Cell death & disease
Yao H,Ren D,Wang Y,Wu L,Wu Y,Wang W,Li Q,Liu L
Fri Sep 02 00:00:00 EDT 2022
Stewed Rhubarb Decoction Ameliorates Adenine-Induced Chronic Renal Failure in Mice by Regulating Gut Microbiota Dysbiosis.
Frontiers in pharmacology
Wang R,Hu B,Ye C,Zhang Z,Yin M,Cao Q,Ba Y,Liu H
Sat Apr 09 00:00:00 EDT 2022
Regulation of ZO-1 on β-catenin mediates sulforaphane suppressed colorectal cancer stem cell properties in colorectal cancer.
Food & function
Chen Y,Tang L,Ye X,Chen Y,Shan E,Han H,Zhong C
Mon Nov 28 00:00:00 EST 2022
Regulation of ZO-1 on β-catenin mediates sulforaphane suppressed colorectal cancer stem cell properties in colorectal cancer.
Food & function
Chen Y,Tang L,Ye X,Chen Y,Shan E,Han H,Zhong C
Mon Nov 28 00:00:00 EST 2022
Regulation of ZO-1 on β-catenin mediates sulforaphane suppressed colorectal cancer stem cell properties in colorectal cancer.
Food & function
Chen Y,Tang L,Ye X,Chen Y,Shan E,Han H,Zhong C
Mon Nov 28 00:00:00 EST 2022
Regulation of ZO-1 on β-catenin mediates sulforaphane suppressed colorectal cancer stem cell properties in colorectal cancer.
Food & function
Chen Y,Tang L,Ye X,Chen Y,Shan E,Han H,Zhong C
Mon Nov 28 00:00:00 EST 2022
G protein subunit gamma 5 promotes the proliferation, metastasis and glycolysis of breast cancer cells through the Wnt/β-catenin pathway.
Anti-cancer drugs
Yuan Z,Ren R,Xu Z
Tue Nov 01 00:00:00 EDT 2022
WNT6 participates in the occurrence and development of ovarian cancer by upregulating/activating the typical Wnt pathway and Notch1 signaling pathway.
Gene
Bao H,Wu W,Li Y,Zong Z,Chen S
Tue Dec 20 00:00:00 EST 2022
Comprehensive Transcriptome Analysis of Hair Follicle Morphogenesis Reveals That lncRNA-H19 Promotes Dermal Papilla Cell Proliferation through the Chi-miR-214-3p/β-Catenin Axis in Cashmere Goats.
International journal of molecular sciences
Zhang Y,Li F,Shi Y,Zhang T,Wang X
Fri Sep 02 00:00:00 EDT 2022
Ginsenoside Rh4 Suppresses Metastasis of Esophageal Cancer and Expression of c-Myc via Targeting the Wnt/β-Catenin Signaling Pathway.
Nutrients
Chen J,Duan Z,Liu Y,Fu R,Zhu C
Mon Jul 25 00:00:00 EDT 2022
The ETS1-LINC00278 negative feedback loop plays a role in COL4A1/COL4A2 regulation in laryngeal squamous cell carcinoma.
Neoplasma
Yang C,Cao H,Yang JW,Wang JT,Yu MM,Wang BS
Fri Jul 01 00:00:00 EDT 2022
The Chinese medicine Fufang Zhenzhu Tiaozhi capsule protects against atherosclerosis by suppressing EndMT via modulating Akt1/β-catenin signaling pathway.
Journal of ethnopharmacology
Diao H,Cheng J,Huang X,Huang B,Shao X,Zhao J,Lan D,Zhu Q,Yan M,Zhang Y,Rong X,Guo J
Fri Jul 15 00:00:00 EDT 2022
SiNiSan alleviates liver injury by promoting hepatic stem cell differentiation via Wnt/β-catenin signaling pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Xu W,Du X,Li J,Zhang Z,Ma X,Luo D,Xiao M,Sun Q
Sun May 01 00:00:00 EDT 2022
SiNiSan alleviates liver injury by promoting hepatic stem cell differentiation via Wnt/β-catenin signaling pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Xu W,Du X,Li J,Zhang Z,Ma X,Luo D,Xiao M,Sun Q
Sun May 01 00:00:00 EDT 2022
Sestrin2 reduces cancer stemness via Wnt/β-catenin signaling in colorectal cancer.
Cancer cell international
Wei J,Zheng X,Li W,Li X,Fu Z
Fri Feb 11 00:00:00 EST 2022
Sestrin2 reduces cancer stemness via Wnt/β-catenin signaling in colorectal cancer.
Cancer cell international
Wei J,Zheng X,Li W,Li X,Fu Z
Fri Feb 11 00:00:00 EST 2022
Sestrin2 reduces cancer stemness via Wnt/β-catenin signaling in colorectal cancer.
Cancer cell international
Wei J,Zheng X,Li W,Li X,Fu Z
Fri Feb 11 00:00:00 EST 2022
Sestrin2 reduces cancer stemness via Wnt/β-catenin signaling in colorectal cancer.
Cancer cell international
Wei J,Zheng X,Li W,Li X,Fu Z
Fri Feb 11 00:00:00 EST 2022
Bergamottin promotes osteoblast differentiation and bone formation via activating the Wnt/β-catenin signaling pathway.
Food & function
Wang X,Tian Y,Liang X,Yin C,Huai Y,Zhao Y,Huang Q,Chu X,Wang W,Qian A
Mon Mar 07 00:00:00 EST 2022
The miR-34a/WNT7B modulates the sensitivity of cholangiocarcinoma cells to p53-mediated photodynamic therapy toxicity.
Biochemical and biophysical research communications
Han Y,Yang Y,Huang S,Yao L,Wu L
Sat Feb 05 00:00:00 EST 2022
Long non-coding RNA BBOX1-AS1 exacerbates esophageal squamous cell carcinoma development by regulating HOXB7/β-catenin axis.
Experimental cell research
Sheng J,Zhou M,Wang C,Jia J,Chu J,Ju C,Wan J,He J,He F
Wed Jun 01 00:00:00 EDT 2022
Identification of KIF4A as a pan-cancer diagnostic and prognostic biomarker via bioinformatics analysis and validation in osteosarcoma cell lines.
PeerJ
Pan J,Lei X,Mao X
Sat Apr 23 00:00:00 EDT 2022
Kochia scoparia Saponin Momordin Ic Modulates HaCaT Cell Proliferation and Apoptosis via the Wnt/β-Catenin Pathway.
Evidence-based complementary and alternative medicine : eCAM
Luo M,Zeng B,Wang H,Yang Z,Peng Y,Zhang Y,Wang C
Sat Jul 31 00:00:00 EDT 2021
Kochia scoparia Saponin Momordin Ic Modulates HaCaT Cell Proliferation and Apoptosis via the Wnt/β-Catenin Pathway.
Evidence-based complementary and alternative medicine : eCAM
Luo M,Zeng B,Wang H,Yang Z,Peng Y,Zhang Y,Wang C
Sat Jul 31 00:00:00 EDT 2021
Gastrin-17 induces gastric cancer cell epithelial-mesenchymal transition via the Wnt/β-catenin signaling pathway.
Journal of physiology and biochemistry
Li Y,Zhao Y,Li Y,Zhang X,Li C,Long N,Chen X,Bao L,Zhou J,Xie Y
Mon Feb 01 00:00:00 EST 2021
Gastrin-17 induces gastric cancer cell epithelial-mesenchymal transition via the Wnt/β-catenin signaling pathway.
Journal of physiology and biochemistry
Li Y,Zhao Y,Li Y,Zhang X,Li C,Long N,Chen X,Bao L,Zhou J,Xie Y
Mon Feb 01 00:00:00 EST 2021
BK Channel Deficiency in Osteoblasts Reduces Bone Formation via the Wnt/β-Catenin Pathway.
Molecules and cells
Jiang L,Yang Q,Gao J,Yang J,He J,Xin H,Zhang X
Tue Aug 31 00:00:00 EDT 2021
BK Channel Deficiency in Osteoblasts Reduces Bone Formation via the Wnt/β-Catenin Pathway.
Molecules and cells
Jiang L,Yang Q,Gao J,Yang J,He J,Xin H,Zhang X
Tue Aug 31 00:00:00 EDT 2021
BK Channel Deficiency in Osteoblasts Reduces Bone Formation via the Wnt/β-Catenin Pathway.
Molecules and cells
Jiang L,Yang Q,Gao J,Yang J,He J,Xin H,Zhang X
Tue Aug 31 00:00:00 EDT 2021
BK Channel Deficiency in Osteoblasts Reduces Bone Formation via the Wnt/β-Catenin Pathway.
Molecules and cells
Jiang L,Yang Q,Gao J,Yang J,He J,Xin H,Zhang X
Tue Aug 31 00:00:00 EDT 2021
Chromobox 4 facilitates tumorigenesis of lung adenocarcinoma through the Wnt/β-catenin pathway.
Neoplasia (New York, N.Y.)
Wang Z,Fang Z,Chen G,Liu B,Xu J,Li F,Li F,Liu H,Zhang H,Sun Y,Tian G,Chen H,Xu G,Zhang L,Hu L,Ji H
Mon Feb 01 00:00:00 EST 2021
Chromobox 4 facilitates tumorigenesis of lung adenocarcinoma through the Wnt/β-catenin pathway.
Neoplasia (New York, N.Y.)
Wang Z,Fang Z,Chen G,Liu B,Xu J,Li F,Li F,Liu H,Zhang H,Sun Y,Tian G,Chen H,Xu G,Zhang L,Hu L,Ji H
Mon Feb 01 00:00:00 EST 2021
Chromobox 4 facilitates tumorigenesis of lung adenocarcinoma through the Wnt/β-catenin pathway.
Neoplasia (New York, N.Y.)
Wang Z,Fang Z,Chen G,Liu B,Xu J,Li F,Li F,Liu H,Zhang H,Sun Y,Tian G,Chen H,Xu G,Zhang L,Hu L,Ji H
Mon Feb 01 00:00:00 EST 2021
Chlorogenic Acid Promotes Osteogenic Differentiation of Human Dental Pulp Stem Cells Through Wnt Signaling.
Stem cells and development
Hu X,Wang L,He Y,Wei M,Yan H,Zhu H
Tue Jun 15 00:00:00 EDT 2021
TRIM11 Promotes Proliferation, Migration, Invasion and EMT of Gastric Cancer by Activating β-Catenin Signaling.
OncoTargets and therapy
Lan Q,Tan X,He P,Li W,Tian S,Dong W
Thu Apr 21 00:00:00 EDT 2022
TRIM11 Promotes Proliferation, Migration, Invasion and EMT of Gastric Cancer by Activating β-Catenin Signaling.
OncoTargets and therapy
Lan Q,Tan X,He P,Li W,Tian S,Dong W
Thu Apr 21 00:00:00 EDT 2022
USP36-Mediated Deubiquitination of DOCK4 Contributes to the Diabetic Renal Tubular Epithelial Cell Injury via Wnt/β-Catenin Signaling Pathway.
Frontiers in cell and developmental biology
Zhu S,Hou S,Lu Y,Sheng W,Cui Z,Dong T,Feng H,Wan Q
Thu May 13 00:00:00 EDT 2021
An SETD1A/Wnt/β-catenin feedback loop promotes NSCLC development.
Journal of experimental & clinical cancer research : CR
Wang R,Liu J,Li K,Yang G,Chen S,Wu J,Xie X,Ren H,Pang Y
Wed Oct 13 00:00:00 EDT 2021
An SETD1A/Wnt/β-catenin feedback loop promotes NSCLC development.
Journal of experimental & clinical cancer research : CR
Wang R,Liu J,Li K,Yang G,Chen S,Wu J,Xie X,Ren H,Pang Y
Wed Oct 13 00:00:00 EDT 2021
An SETD1A/Wnt/β-catenin feedback loop promotes NSCLC development.
Journal of experimental & clinical cancer research : CR
Wang R,Liu J,Li K,Yang G,Chen S,Wu J,Xie X,Ren H,Pang Y
Wed Oct 13 00:00:00 EDT 2021
An SETD1A/Wnt/β-catenin feedback loop promotes NSCLC development.
Journal of experimental & clinical cancer research : CR
Wang R,Liu J,Li K,Yang G,Chen S,Wu J,Xie X,Ren H,Pang Y
Wed Oct 13 00:00:00 EDT 2021
Sini San Inhibits Chronic Psychological Stress-Induced Breast Cancer Stemness by Suppressing Cortisol-Mediated GRP78 Activation.
Frontiers in pharmacology
Zheng Y,Zhang J,Huang W,Zhong LLD,Wang N,Wang S,Yang B,Wang X,Pan B,Situ H,Lin Y,Liu X,Shi Y,Wang Z
Fri Dec 17 00:00:00 EST 2021
Knockdown of USP9X reverses cisplatin resistance by decreasing β-catenin expression in nasopharyngeal carcinoma cells.
Neoplasma
Lu WX,Gan GS,Yang B
Thu Jul 01 00:00:00 EDT 2021
Knockdown of USP9X reverses cisplatin resistance by decreasing β-catenin expression in nasopharyngeal carcinoma cells.
Neoplasma
Lu WX,Gan GS,Yang B
Thu Jul 01 00:00:00 EDT 2021
Membrane-cytoplasm translocation of annexin A4 is involved in the metastasis of colorectal carcinoma.
Aging
Peng Y,Zhang Z,Zhang A,Liu C,Sun Y,Peng Z,Liu Y
Wed Mar 24 00:00:00 EDT 2021
Paired-like homeodomain transcription factor 2 affects endometrial cell function and embryo implantation through the Wnt/β-catenin pathway.
Cell biology international
Zhang H,Qi J,Guo J,Wang Y,Guan Y,Fan J,Sui L,Xu Y,Kong L,Yan B,Kong Y
Wed Sep 01 00:00:00 EDT 2021
Bromodomain-containing protein 9 promotes hepatocellular carcinoma progression via activating the Wnt/β-catenin signaling pathway.
Experimental cell research
Fang D,Wang MR,Guan JL,Han YY,Sheng JQ,Tian DA,Li PY
Wed Sep 15 00:00:00 EDT 2021
Tunicamycin-Induced Endoplasmic Reticulum Stress Promotes Breast Cancer Cell MDA-MB-231 Apoptosis through Inhibiting Wnt/β-Catenin Signaling Pathway.
Journal of healthcare engineering
You Z,He L,Yan N
Mon Feb 21 00:00:00 EST 2022
regulates pathological cardiac hypertrophy via a β-catenin-dependent mechanism.
American journal of physiology. Heart and circulatory physiology
Lin L,Xu W,Li Y,Zhu P,Yuan W,Liu M,Shi Y,Chen Y,Liang J,Chen J,Yang B,Cai W,Wen Y,Zhu X,Peng X,Zhou Z,Mo X,Wan Y,Yuan H,Li F,Ye X,Jiang Z,Wang Y,Zhuang J,Fan X,Wu X
Thu Apr 01 00:00:00 EDT 2021
LRRC17 regulates the bone metabolism of human bone marrow mesenchymal stem cells from patients with idiopathic necrosis of femoral head through Wnt signaling pathways: A preliminary report.
Experimental and therapeutic medicine
Song D,Wu ZS,Xu Q,Wang K,Xu MT,Ha CZ,Zhang C,Wang DW
Thu Jul 01 00:00:00 EDT 2021
RTEF-1 Inhibits Vascular Smooth Muscle Cell Calcification through Regulating Wnt/β-Catenin Signaling Pathway.
Calcified tissue international
Cong J,Cheng B,Liu J,He P
Sun Aug 01 00:00:00 EDT 2021
DNA methylation mediated RSPO2 to promote follicular development in mammals.
Cell death & disease
Zhou X,He Y,Li N,Bai G,Pan X,Zhang Z,Zhang H,Li J,Yuan X
Sat Jun 26 00:00:00 EDT 2021
ZEB1 Mediates Bone Marrow Mesenchymal Stem Cell Osteogenic Differentiation Partly via Wnt/β-Catenin Signaling.
Frontiers in molecular biosciences
Xu C,Shi H,Jiang X,Fan Y,Huang D,Qi X,Cheng Q
Sat Jun 12 00:00:00 EDT 2021
Compound Kushen Injection intervenes metabolic reprogramming and epithelial-mesenchymal transition of HCC via regulating β-catenin/c-Myc signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang KX,Du GH,Qin XM,Gao L
Wed Dec 01 00:00:00 EST 2021
Compound Kushen Injection intervenes metabolic reprogramming and epithelial-mesenchymal transition of HCC via regulating β-catenin/c-Myc signaling.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang KX,Du GH,Qin XM,Gao L
Wed Dec 01 00:00:00 EST 2021
Silencing Proteasome 26S Subunit ATPase 2 (PSMC2) Protects the Osteogenic Differentiation In Vitro and Osteogenesis In Vivo.
Calcified tissue international
Zhou B,Peng K,Wang G,Chen W,Kang Y
Thu Jul 01 00:00:00 EDT 2021
Silencing Proteasome 26S Subunit ATPase 2 (PSMC2) Protects the Osteogenic Differentiation In Vitro and Osteogenesis In Vivo.
Calcified tissue international
Zhou B,Peng K,Wang G,Chen W,Kang Y
Thu Jul 01 00:00:00 EDT 2021
Polo Like Kinase 4 (PLK4) impairs human bone marrow mesenchymal stem cell (BMSC) viability and osteogenic differentiation.
Biochemical and biophysical research communications
Zhou B,Peng K,Wang G,Chen W,Kang Y
Fri Apr 16 00:00:00 EDT 2021
TM9SF4 is a novel regulator in lineage commitment of bone marrow mesenchymal stem cells to either osteoblasts or adipocytes.
Stem cell research & therapy
Yu L,Xie M,Zhang F,Wan C,Yao X
Sat Nov 13 00:00:00 EST 2021
DNMT1-Induced miR-152-3p Suppression Facilitates Cardiac Fibroblast Activation in Cardiac Fibrosis.
Cardiovascular toxicology
Xu SS,Ding JF,Shi P,Shi KH,Tao H
Wed Dec 01 00:00:00 EST 2021
POSTN Promotes the Proliferation of Spermatogonial Cells by Activating the Wnt/β-Catenin Signaling Pathway.
Reproductive sciences (Thousand Oaks, Calif.)
Li C,Cheng D,Xu P,Nie H,Zhang T,Pang X
Fri Oct 01 00:00:00 EDT 2021
Oxidative Stress-Induced Hypermethylation of KLF5 Promoter Mediated by DNMT3B Impairs Osteogenesis by Diminishing the Interaction with β-Catenin.
Antioxidants & redox signaling
Li L,Wang H,Chen X,Li X,Wang G,Jie Z,Zhao X,Sun X,Huang H,Fan S,Xie Z,Wang J
Thu Jul 01 00:00:00 EDT 2021
Enriched environment remedies cognitive dysfunctions and synaptic plasticity through NMDAR-Ca2+-Activin A circuit in chronic cerebral hypoperfusion rats.
Aging
Zhang X,Shi X,Wang J,Xu Z,He J
Mon Aug 30 00:00:00 EDT 2021
Scutellarin ameliorates colitis-associated colorectal cancer by suppressing Wnt/β-catenin signaling cascade.
European journal of pharmacology
Zeng S,Chen L,Sun Q,Zhao H,Yang H,Ren S,Liu M,Meng X,Xu H
Sun Sep 05 00:00:00 EDT 2021
Scutellarin ameliorates colitis-associated colorectal cancer by suppressing Wnt/β-catenin signaling cascade.
European journal of pharmacology
Zeng S,Chen L,Sun Q,Zhao H,Yang H,Ren S,Liu M,Meng X,Xu H
Sun Sep 05 00:00:00 EDT 2021
Scutellarin ameliorates colitis-associated colorectal cancer by suppressing Wnt/β-catenin signaling cascade.
European journal of pharmacology
Zeng S,Chen L,Sun Q,Zhao H,Yang H,Ren S,Liu M,Meng X,Xu H
Sun Sep 05 00:00:00 EDT 2021
Scutellarin ameliorates colitis-associated colorectal cancer by suppressing Wnt/β-catenin signaling cascade.
European journal of pharmacology
Zeng S,Chen L,Sun Q,Zhao H,Yang H,Ren S,Liu M,Meng X,Xu H
Sun Sep 05 00:00:00 EDT 2021
Scutellarin ameliorates colitis-associated colorectal cancer by suppressing Wnt/β-catenin signaling cascade.
European journal of pharmacology
Zeng S,Chen L,Sun Q,Zhao H,Yang H,Ren S,Liu M,Meng X,Xu H
Sun Sep 05 00:00:00 EDT 2021
Scutellarin ameliorates colitis-associated colorectal cancer by suppressing Wnt/β-catenin signaling cascade.
European journal of pharmacology
Zeng S,Chen L,Sun Q,Zhao H,Yang H,Ren S,Liu M,Meng X,Xu H
Sun Sep 05 00:00:00 EDT 2021
DEK is highly expressed in breast cancer and is associated with malignant phenotype and progression.
Oncology letters
Yang MQ,Bai LL,Wang Z,Lei L,Zheng YW,Li ZH,Huang WJ,Liu CC,Xu HT
Tue Jun 01 00:00:00 EDT 2021
Circ_0119872 promotes uveal melanoma development by regulating the miR-622/G3BP1 axis and downstream signalling pathways.
Journal of experimental & clinical cancer research : CR
Liu S,Chen L,Chen H,Xu K,Peng X,Zhang M
Fri Feb 12 00:00:00 EST 2021
Integrated Strategy of Network Pharmacological Prediction and Experimental Validation Elucidate Possible Mechanism of Bu-Yang Herbs in Treating Postmenopausal Osteoporosis via ESR1.
Frontiers in pharmacology
Xia H,Liu J,Yang W,Liu M,Luo Y,Yang Z,Xie J,Zeng H,Xu R,Ling H,Zeng Q,Xu H,Fang L,Wang H,Tong P,Jin H,Yang F
Sun May 30 00:00:00 EDT 2021
Primary cilia regulate gastric cancer-induced bone loss via cilia/Wnt/β-catenin signaling pathway.
Aging
Xu J,Deng X,Wu X,Zhu H,Zhu Y,Liu J,Chen Q,Yuan C,Liu G,Wang C
Tue Mar 09 00:00:00 EST 2021
Long noncoding RNA MEG3 expressed in human dental pulp regulates LPS-Induced inflammation and odontogenic differentiation in pulpitis.
Experimental cell research
Liu M,Chen L,Wu J,Lin Z,Huang S
Mon Mar 15 00:00:00 EDT 2021
UM-6 induces autophagy and apoptosis via the Hippo-YAP signaling pathway in cervical cancer.
Cancer letters
Wang D,He J,Dong J,Wu S,Liu S,Zhu H,Xu T
Thu Oct 28 00:00:00 EDT 2021
TMEM98, a novel secretory protein, promotes endothelial cell adhesion as well as vascular smooth muscle cell proliferation and migration.
Canadian journal of physiology and pharmacology
Li M,Zhu H,Hu X,Gao F,Hu X,Cui Y,Wei X,Xie C,Lv G,Zhao Y,Gao Y
Sat May 01 00:00:00 EDT 2021
Knockdown of ST6Gal-I expression in human hepatocellular carcinoma cells inhibits their exosome-mediated proliferation- and migration-promoting effects.
IUBMB life
Wang L,Chen X,Wang L,Wang S,Li W,Liu Y,Zhang J
Mon Nov 01 00:00:00 EDT 2021
miR-378d suppresses malignant phenotype of ESCC cells through AKT signaling.
Cancer cell international
Peng J,Shi S,Yu J,Liu J,Wei H,Song H,Wang S,Li Z,He S,Li L,Zhang H,Yan Z,Zhao R,Liu Y,Liu Y,Li J,Zhang R,Wang W
Wed Dec 22 00:00:00 EST 2021
c-Myc Upregulated by High Glucose Inhibits HaCaT Differentiation by S100A6 Transcriptional Activation.
Frontiers in endocrinology
Zhang J,Yang P,Liu D,Gao M,Wang J,Wang X,Liu Y,Zhang X
Wed Dec 22 00:00:00 EST 2021
The ALDH Family Contributes to Immunocyte Infiltration, Proliferation and Epithelial-Mesenchymal Transformation in Glioma.
Frontiers in immunology
Wang Z,Mo Y,Tan Y,Wen Z,Dai Z,Zhang H,Zhang X,Feng S,Liang X,Song T,Cheng Q
Mon Feb 28 00:00:00 EST 2022
Human placental-derived stem cell therapy ameliorates experimental necrotizing enterocolitis.
American journal of physiology. Gastrointestinal and liver physiology
Weis VG,Deal AC,Mekkey G,Clouse C,Gaffley M,Whitaker E,Peeler CB,Weis JA,Schwartz MZ,Atala A
Thu Apr 01 00:00:00 EDT 2021
p53-targeted lncRNA ST7-AS1 acts as a tumour suppressor by interacting with PTBP1 to suppress the Wnt/β-catenin signalling pathway in glioma.
Cancer letters
Sheng J,He X,Yu W,Chen Y,Long Y,Wang K,Zhu S,Liu Q
Sat Apr 10 00:00:00 EDT 2021
p53-targeted lncRNA ST7-AS1 acts as a tumour suppressor by interacting with PTBP1 to suppress the Wnt/β-catenin signalling pathway in glioma.
Cancer letters
Sheng J,He X,Yu W,Chen Y,Long Y,Wang K,Zhu S,Liu Q
Sat Apr 10 00:00:00 EDT 2021
Roles of WNT6 in Sheep Endometrial Epithelial Cell Cycle Progression and Uterine Glands Organogenesis.
Veterinary sciences
Gao X,Yao X,Li X,Liang Y,Liu Z,Wang Z,Li K,Li Y,Zhang G,Wang F
Thu Dec 09 00:00:00 EST 2021
LGR5 induces β-catenin activation and augments tumour progression by activating STAT3 in human intrahepatic cholangiocarcinoma.
Liver international : official journal of the International Association for the Study of the Liver
Kawasaki K,Kuboki S,Furukawa K,Takayashiki T,Takano S,Ohtsuka M
Thu Apr 01 00:00:00 EDT 2021
LGR5 induces β-catenin activation and augments tumour progression by activating STAT3 in human intrahepatic cholangiocarcinoma.
Liver international : official journal of the International Association for the Study of the Liver
Kawasaki K,Kuboki S,Furukawa K,Takayashiki T,Takano S,Ohtsuka M
Thu Apr 01 00:00:00 EDT 2021
Multi-omics analysis to identify susceptibility genes for colorectal cancer.
Human molecular genetics
Yuan Y,Bao J,Chen Z,Villanueva AD,Wen W,Wang F,Zhao D,Fu X,Cai Q,Long J,Shu XO,Zheng D,Moreno V,Zheng W,Lin W,Guo X
Tue Apr 27 00:00:00 EDT 2021
EphA2 super-enhancer promotes tumor progression by recruiting FOSL2 and TCF7L2 to activate the target gene EphA2.
Cell death & disease
Cui S,Wu Q,Liu M,Su M,Liu S,Shao L,Han X,He H
Fri Mar 12 00:00:00 EST 2021
The Role of miR-640: A Potential Suppressor in Breast Cancer via Wnt7b/β-catenin Signaling Pathway.
Frontiers in oncology
Tang C,Wang X,Ji C,Zheng W,Yu Y,Deng X,Zhou X,Fang L
Fri Apr 30 00:00:00 EDT 2021
Polychlorinated biphenyl quinone induced the acquisition of cancer stem cells properties and epithelial-mesenchymal transition through Wnt/β-catenin.
Chemosphere
Qin Q,Yang B,Liu Z,Xu L,Song E,Song Y
Fri Jan 01 00:00:00 EST 2021
Polychlorinated biphenyl quinone induced the acquisition of cancer stem cells properties and epithelial-mesenchymal transition through Wnt/β-catenin.
Chemosphere
Qin Q,Yang B,Liu Z,Xu L,Song E,Song Y
Fri Jan 01 00:00:00 EST 2021
Dihydroartemisinin attenuates osteoarthritis by inhibiting abnormal bone remodeling and angiogenesis in subchondral bone.
International journal of molecular medicine
Ma L,Zhao X,Liu Y,Wu J,Yang X,Jin Q
Mon Mar 01 00:00:00 EST 2021
Autophagy-Induced HDAC6 Activity During Hypoxia Regulates Mitochondrial Energy Metabolism Through the β-Catenin/COUP-TFII Axis in Hepatocellular Carcinoma Cells.
Frontiers in oncology
Yan X,Qu X,Liu B,Zhao Y,Xu L,Yu S,Wang J,Wang L,Su J
Tue Dec 07 00:00:00 EST 2021
SHARPIN stabilizes β-catenin through a linear ubiquitination-independent manner to support gastric tumorigenesis.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association
Zhang L,Liu Q,Liu KW,Qin ZY,Zhu GX,Shen LT,Zhang N,Liu BY,Che LR,Li JY,Wang T,Wen LZ,Liu KJ,Guo Y,Yin XR,Wang XW,Zhou ZH,Xiao HL,Cui YH,Bian XW,Lan CH,Chen D,Wang B
Mon Mar 01 00:00:00 EST 2021
BML-111, the lipoxin A4 agonist, modulates VEGF or CoCl2-induced migration, angiogenesis and permeability in tumor-derived endothelial cells.
Immunology letters
Lin L,Wang Q,Xu F,Luo X,Xu J,Yan L,Li Q,Hao H
Mon Feb 01 00:00:00 EST 2021
BML-111, the lipoxin A4 agonist, modulates VEGF or CoCl2-induced migration, angiogenesis and permeability in tumor-derived endothelial cells.
Immunology letters
Lin L,Wang Q,Xu F,Luo X,Xu J,Yan L,Li Q,Hao H
Mon Feb 01 00:00:00 EST 2021
Apatinib suppresses lung cancer stem-like cells by complex interplay between β-catenin signaling and mitochondrial ROS accumulation.
Cell death discovery
Zhu J,Li X,Liang C,Zhou X,Ge M,Chen Y,Jin J,Yin J,Xu H,Xie C,Zhong C
Wed May 12 00:00:00 EDT 2021
Apatinib suppresses lung cancer stem-like cells by complex interplay between β-catenin signaling and mitochondrial ROS accumulation.
Cell death discovery
Zhu J,Li X,Liang C,Zhou X,Ge M,Chen Y,Jin J,Yin J,Xu H,Xie C,Zhong C
Wed May 12 00:00:00 EDT 2021
Apatinib suppresses lung cancer stem-like cells by complex interplay between β-catenin signaling and mitochondrial ROS accumulation.
Cell death discovery
Zhu J,Li X,Liang C,Zhou X,Ge M,Chen Y,Jin J,Yin J,Xu H,Xie C,Zhong C
Wed May 12 00:00:00 EDT 2021
TSPAN1 promotes autophagy flux and mediates cooperation between WNT-CTNNB1 signaling and autophagy via the MIR454-FAM83A-TSPAN1 axis in pancreatic cancer.
Autophagy
Zhou C,Liang Y,Zhou L,Yan Y,Liu N,Zhang R,Huang Y,Wang M,Tang Y,Ali DW,Wang Y,Michalak M,Chen XZ,Tang J
Fri Oct 01 00:00:00 EDT 2021
TSPAN1 promotes autophagy flux and mediates cooperation between WNT-CTNNB1 signaling and autophagy via the MIR454-FAM83A-TSPAN1 axis in pancreatic cancer.
Autophagy
Zhou C,Liang Y,Zhou L,Yan Y,Liu N,Zhang R,Huang Y,Wang M,Tang Y,Ali DW,Wang Y,Michalak M,Chen XZ,Tang J
Fri Oct 01 00:00:00 EDT 2021
Cancer-associated fibroblast-derived SDF-1 induces epithelial-mesenchymal transition of lung adenocarcinoma via CXCR4/β-catenin/PPARδ signalling.
Cell death & disease
Wang Y,Lan W,Xu M,Song J,Mao J,Li C,Du X,Jiang Y,Li E,Zhang R,Wang Q
Fri Feb 26 00:00:00 EST 2021
Condensation tendency and planar isotropic actin gradient induce radial alignment in confined monolayers.
eLife
Xie T,St Pierre SR,Olaranont N,Brown LE,Wu M,Sun Y
Mon Sep 20 00:00:00 EDT 2021
Liposomal honokiol promotes hair growth via activating Wnt3a/β-catenin signaling pathway and down regulating TGF-β1 in C57BL/6N mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li S,Chen J,Chen F,Wang C,Guo X,Wang C,Fan Y,Wang Y,Peng Y,Li W
Wed Sep 01 00:00:00 EDT 2021
Liposomal honokiol promotes hair growth via activating Wnt3a/β-catenin signaling pathway and down regulating TGF-β1 in C57BL/6N mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li S,Chen J,Chen F,Wang C,Guo X,Wang C,Fan Y,Wang Y,Peng Y,Li W
Wed Sep 01 00:00:00 EDT 2021
Liposomal honokiol promotes hair growth via activating Wnt3a/β-catenin signaling pathway and down regulating TGF-β1 in C57BL/6N mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Li S,Chen J,Chen F,Wang C,Guo X,Wang C,Fan Y,Wang Y,Peng Y,Li W
Wed Sep 01 00:00:00 EDT 2021
The protective mechanism of SIRT1 on cartilage through regulation of LEF-1.
BMC musculoskeletal disorders
Hu X,Feng G,Meng Z,Ma L,Jin Q
Tue Jul 27 00:00:00 EDT 2021
Targeting ADAR1 suppresses progression and peritoneal metastasis of gastric cancer through Wnt / β-catenin pathway.
Journal of Cancer
Li Z,Huang Y,Xu Y,Wang X,Wang H,Zhao S,Liu H,Yu G,Che X
Sat Feb 19 00:00:00 EST 2022
Targeting ADAR1 suppresses progression and peritoneal metastasis of gastric cancer through Wnt / β-catenin pathway.
Journal of Cancer
Li Z,Huang Y,Xu Y,Wang X,Wang H,Zhao S,Liu H,Yu G,Che X
Sat Feb 19 00:00:00 EST 2022
Targeting ADAR1 suppresses progression and peritoneal metastasis of gastric cancer through Wnt / β-catenin pathway.
Journal of Cancer
Li Z,Huang Y,Xu Y,Wang X,Wang H,Zhao S,Liu H,Yu G,Che X
Sat Feb 19 00:00:00 EST 2022
MCC Regulator of WNT Signaling Pathway (MCC) Is a Podocyte Essential Gene.
Frontiers in medicine
Song H,Zhuang L,Xu X,Shi J,Hu W,Liu Z,Shi S
Tue Dec 21 00:00:00 EST 2021
LncRNA MIR155HG Promotes Temozolomide Resistance by Activating the Wnt/β-Catenin Pathway Via Binding to PTBP1 in Glioma.
Cellular and molecular neurobiology
He X,Sheng J,Yu W,Wang K,Zhu S,Liu Q
Sun Aug 01 00:00:00 EDT 2021
LncRNA MIR155HG Promotes Temozolomide Resistance by Activating the Wnt/β-Catenin Pathway Via Binding to PTBP1 in Glioma.
Cellular and molecular neurobiology
He X,Sheng J,Yu W,Wang K,Zhu S,Liu Q
Sun Aug 01 00:00:00 EDT 2021
Protocatechuic aldehyde protects cardiomycoytes against ischemic injury via regulation of nuclear pyruvate kinase M2.
Acta pharmaceutica Sinica. B
Wu X,Liu L,Zheng Q,Hao H,Ye H,Li P,Yang H
Mon Nov 01 00:00:00 EDT 2021
CCT8 recovers WTp53-suppressed cell cycle evolution and EMT to promote colorectal cancer progression.
Oncogenesis
Liao Q,Ren Y,Yang Y,Zhu X,Zhi Y,Zhang Y,Chen Y,Ding Y,Zhao L
Fri Dec 03 00:00:00 EST 2021
Investigation for GSK3β expression in diabetic osteoporosis and negative osteogenic effects of GSK3β on bone marrow mesenchymal stem cells under a high glucose microenvironment.
Biochemical and biophysical research communications
Chen Y,Chen L,Huang R,Yang W,Chen S,Lin K,Liu J
Fri Jan 01 00:00:00 EST 2021
Investigation for GSK3β expression in diabetic osteoporosis and negative osteogenic effects of GSK3β on bone marrow mesenchymal stem cells under a high glucose microenvironment.
Biochemical and biophysical research communications
Chen Y,Chen L,Huang R,Yang W,Chen S,Lin K,Liu J
Fri Jan 01 00:00:00 EST 2021
Inhibition of growth and metastasis of breast cancer by targeted delivery of 17-hydroxy-jolkinolide B via hyaluronic acid-coated liposomes.
Carbohydrate polymers
Liu D,Zhang Q,Wang J,Guan S,Cai D,Liu J
Thu Apr 01 00:00:00 EDT 2021
AXIN2 Reduces the Survival of Porcine Induced Pluripotent Stem Cells (piPSCs).
International journal of molecular sciences
Zhang R,Yu S,Shen Q,Zhao W,Zhang J,Wu X,Zhu Z,Wu X,Li N,Peng S,Hua J
Tue Nov 30 00:00:00 EST 2021
AXIN2 Reduces the Survival of Porcine Induced Pluripotent Stem Cells (piPSCs).
International journal of molecular sciences
Zhang R,Yu S,Shen Q,Zhao W,Zhang J,Wu X,Zhu Z,Wu X,Li N,Peng S,Hua J
Tue Nov 30 00:00:00 EST 2021
Long noncoding RNA LYPLAL1-AS1 regulates adipogenic differentiation of human mesenchymal stem cells by targeting desmoplakin and inhibiting the Wnt/β-catenin pathway.
Cell death discovery
Yang Y,Fan J,Xu H,Fan L,Deng L,Li J,Li D,Li H,Zhang F,Zhao RC
Sat May 15 00:00:00 EDT 2021
Expression of TCF3 in Wilms' tumor and its regulatory role in kidney tumor cell viability, migration and apoptosis in vitro.
Molecular medicine reports
Zhou N,Yan B,Ma J,Jiang H,Li L,Tang H,Ji F,Yao Z
Wed Sep 01 00:00:00 EDT 2021
miR-1246 shuttling from fibroblasts promotes colorectal cancer cell migration.
Neoplasma
Si G,Li S,Zheng Q,Zhu S,Zhou C
Mon Mar 01 00:00:00 EST 2021
miR-1246 shuttling from fibroblasts promotes colorectal cancer cell migration.
Neoplasma
Si G,Li S,Zheng Q,Zhu S,Zhou C
Mon Mar 01 00:00:00 EST 2021
NEK2 plays an active role in Tumorigenesis and Tumor Microenvironment in Non-Small Cell Lung Cancer.
International journal of biological sciences
Bai R,Yuan C,Sun W,Zhang J,Luo Y,Gao Y,Li Y,Gong Y,Xie C
Fri Mar 25 00:00:00 EDT 2022
α-Klotho released from HK-2 cells inhibits osteogenic differentiation of renal interstitial fibroblasts by inactivating the Wnt-β-catenin pathway.
Cellular and molecular life sciences : CMLS
Zhu Z,Ruan S,Jiang Y,Huang F,Xia W,Chen J,Cui Y,He C,Zeng F,Li Y,Chen Z,Chen H
Wed Dec 01 00:00:00 EST 2021
Citrus Alkaline Extracts Inhibit Senescence of A549 Cells to Alleviate Pulmonary Fibrosis via the β-Catenin/P53 Pathway.
Medical science monitor : international medical journal of experimental and clinical research
Han D,Xu Y,Peng WP,Feng F,Wang Z,Gu C,Zhou X
Fri Mar 12 00:00:00 EST 2021
Pyrvinium pamoate can overcome artemisinin's resistance in anaplastic thyroid cancer.
BMC complementary medicine and therapies
Li Y
Fri May 28 00:00:00 EDT 2021
Specific deletion of protein phosphatase 6 catalytic subunit in Sertoli cells leads to disruption of spermatogenesis.
Cell death & disease
Lei WL,Li YY,Meng TG,Ning Y,Sun SM,Zhang CH,Gui Y,Wang ZB,Qian WP,Sun QY
Mon Sep 27 00:00:00 EDT 2021
Specific deletion of protein phosphatase 6 catalytic subunit in Sertoli cells leads to disruption of spermatogenesis.
Cell death & disease
Lei WL,Li YY,Meng TG,Ning Y,Sun SM,Zhang CH,Gui Y,Wang ZB,Qian WP,Sun QY
Mon Sep 27 00:00:00 EDT 2021
LEM domain containing 1 promotes thyroid cancer cell proliferation and migration by activating the Wnt/β-catenin signaling pathway and epithelial-mesenchymal transition.
Oncology letters
Xu M,Lin B,Zheng D,Wen J,Hu W,Li C,Zhang X,Zhang X,Qu J
Tue Jun 01 00:00:00 EDT 2021
miR-937-5p targets SOX17 to modulate breast cancer cell cycle and cell proliferation through the Wnt signaling pathway.
Cellular signalling
Xiong X,Xu W,Gong J,Wang L,Dai M,Chen G,Yuan L
Fri Jan 01 00:00:00 EST 2021
Interaction of FLNA and ANXA2 promotes gefitinib resistance by activating the Wnt pathway in non-small-cell lung cancer.
Molecular and cellular biochemistry
Cheng L,Tong Q
Fri Oct 01 00:00:00 EDT 2021
High-mobility group AT-Hook 1 mediates the role of nuclear factor I/X in osteogenic differentiation through activating canonical Wnt signaling.
Stem cells (Dayton, Ohio)
Wu X,Wang X,Shan L,Zhou J,Zhang X,Zhu E,Yuan H,Wang B
Fri Oct 01 00:00:00 EDT 2021
Oleanolic acid inhibits the migration and invasion of hepatocellular carcinoma cells by promoting microRNA-122 expression.
Die Pharmazie
He Y,Liu X,Huang M,Wei Z,Zhang M,He M,Zheng Z,Dong H,Liu D
Wed Sep 01 00:00:00 EDT 2021
Oleanolic acid inhibits the migration and invasion of hepatocellular carcinoma cells by promoting microRNA-122 expression.
Die Pharmazie
He Y,Liu X,Huang M,Wei Z,Zhang M,He M,Zheng Z,Dong H,Liu D
Wed Sep 01 00:00:00 EDT 2021
Artemisia argyi Essential Oil Inhibits Hepatocellular Carcinoma Metastasis via Suppression of DEPDC1 Dependent Wnt/β-Catenin Signaling Pathway.
Frontiers in cell and developmental biology
Li Y,Tian Y,Zhong W,Wang N,Wang Y,Zhang Y,Zhang Z,Li J,Ma F,Zhao Z,Peng Y
Sat Jul 17 00:00:00 EDT 2021
Artemisia argyi Essential Oil Inhibits Hepatocellular Carcinoma Metastasis via Suppression of DEPDC1 Dependent Wnt/β-Catenin Signaling Pathway.
Frontiers in cell and developmental biology
Li Y,Tian Y,Zhong W,Wang N,Wang Y,Zhang Y,Zhang Z,Li J,Ma F,Zhao Z,Peng Y
Sat Jul 17 00:00:00 EDT 2021
The Presence of Circulating Tumor Cell Cluster Characterizes an Aggressive Hepatocellular Carcinoma Subtype.
Frontiers in oncology
Yu JJ,Shu C,Yang HY,Huang Z,Li YN,Tao R,Chen YY,Chen Q,Chen XP,Xiao W
Wed Apr 27 00:00:00 EDT 2022
Overexpression of NCAPG inhibits cardia adenocarcinoma apoptosis and promotes epithelial-mesenchymal transition through the Wnt/β-catenin signaling pathway.
Gene
Zhang X,Zhu M,Wang H,Song Z,Zhan D,Cao W,Han Y,Jia J
Fri Jan 15 00:00:00 EST 2021
A 2-Benzylmalonate Derivative as STAT3 Inhibitor Suppresses Tumor Growth in Hepatocellular Carcinoma by Upregulating β-TrCP E3 Ubiquitin Ligase.
International journal of molecular sciences
Peng T,Wonganan O,Zhang Z,Yu J,Xi R,Cao Y,Suksamrarn A,Zhang G,Wang F
Thu Mar 25 00:00:00 EDT 2021
A 2-Benzylmalonate Derivative as STAT3 Inhibitor Suppresses Tumor Growth in Hepatocellular Carcinoma by Upregulating β-TrCP E3 Ubiquitin Ligase.
International journal of molecular sciences
Peng T,Wonganan O,Zhang Z,Yu J,Xi R,Cao Y,Suksamrarn A,Zhang G,Wang F
Thu Mar 25 00:00:00 EDT 2021
CRISPR/Cas9 library screening uncovered methylated PKP2 as a critical driver of lung cancer radioresistance by stabilizing β-catenin.
Oncogene
Cheng C,Pei X,Li SW,Yang J,Li C,Tang J,Hu K,Huang G,Min WP,Sang Y
Thu Apr 01 00:00:00 EDT 2021
MMP12 is a potential therapeutic target for Adamantinomatous craniopharyngioma: Conclusions from bioinformatics analysis and in vitro experiments.
Oncology letters
Li M,Zhou L,Li S,Fang L,Yang L,Wu X,Yang C,Bao Y,Lan S,Tong Z,Zheng S,Tang B,Zeng E,Xie S,Chen C,Hong T
Thu Jul 01 00:00:00 EDT 2021
Brain Microvascular Endothelial Cell-Derived HMGB1 Facilitates Monocyte Adhesion and Transmigration to Promote JEV Neuroinvasion.
Frontiers in cellular and infection microbiology
Zou SS,Zou QC,Xiong WJ,Cui NY,Wang K,Liu HX,Lou WJ,Higazy D,Zhang YG,Cui M
Tue Oct 19 00:00:00 EDT 2021
Hsa_circ_0001666 suppresses the progression of colorectal cancer through the miR-576-5p/PCDH10 axis.
Clinical and translational medicine
Zhou J,Wang L,Sun Q,Chen R,Zhang C,Yang P,Tan Y,Peng C,Wang T,Jin C,Ji J,Jin K,Sun Y
Mon Nov 01 00:00:00 EDT 2021
TACR2 is associated with the immune microenvironment and inhibits migration and proliferation via the Wnt/β-catenin signaling pathway in prostate cancer.
Cancer cell international
Jianfeng W,Yutao W,Jianbin B
Sat Aug 07 00:00:00 EDT 2021
Transcriptome Differences in Pig Tracheal Epithelial Cells in Response to Pasteurella Multocida Infection.
Frontiers in veterinary science
Wang X,Wang F,Lin L,Liang W,Liu S,Hua L,Wang X,Chen H,Peng Z,Wu B
Wed Sep 08 00:00:00 EDT 2021
Keratin 17 upregulation promotes cell metastasis and angiogenesis in colon adenocarcinoma.
Bioengineered
Ji R,Ji Y,Ma L,Ge S,Chen J,Wu S,Huang T,Sheng Y,Wang L,Yi N,Liu Z
Wed Dec 01 00:00:00 EST 2021
Keratin 17 upregulation promotes cell metastasis and angiogenesis in colon adenocarcinoma.
Bioengineered
Ji R,Ji Y,Ma L,Ge S,Chen J,Wu S,Huang T,Sheng Y,Wang L,Yi N,Liu Z
Wed Dec 01 00:00:00 EST 2021
miR-139-5p Loss-Mediated WTAP Activation Contributes to Hepatocellular Carcinoma Progression by Promoting the Epithelial to Mesenchymal Transition.
Frontiers in oncology
Liu W,Gao X,Chen X,Zhao N,Sun Y,Zou Y,Guan Y,Yang L,Pei X,Wang G,Wang B,Li M,Song W
Tue May 04 00:00:00 EDT 2021
miR-139-5p Loss-Mediated WTAP Activation Contributes to Hepatocellular Carcinoma Progression by Promoting the Epithelial to Mesenchymal Transition.
Frontiers in oncology
Liu W,Gao X,Chen X,Zhao N,Sun Y,Zou Y,Guan Y,Yang L,Pei X,Wang G,Wang B,Li M,Song W
Tue May 04 00:00:00 EDT 2021
Circβ-catenin promotes tumor growth and Warburg effect of gallbladder cancer by regulating STMN1 expression.
Cell death discovery
Wang S,Su T,Tong H,Zhou D,Ma F,Ding J,Hao Y,Shi W,Quan Z
Mon Sep 06 00:00:00 EDT 2021
B2M gene expression shapes the immune landscape of lung adenocarcinoma and determines the response to immunotherapy.
Immunology
Zhao Y,Cao Y,Chen Y,Wu L,Hang H,Jiang C,Zhou X
Mon Nov 01 00:00:00 EDT 2021
Human splenic TER cells: A relevant prognostic factor acting via the artemin-GFRα3-ERK pathway in pancreatic ductal adenocarcinoma.
International journal of cancer
Li TJ,Li H,Zhang WH,Xu SS,Jiang W,Li S,Gao HL,Han X,Xu HX,Wu CT,Wang WQ,Yu XJ,Liu L
Thu Apr 01 00:00:00 EDT 2021
Long Non-Coding RNA CCAT2 Activates RAB14 and Acts as an Oncogene in Colorectal Cancer.
Frontiers in oncology
Wang D,Li Z,Yin H
Tue Dec 07 00:00:00 EST 2021
A Green and Blue Monochromatic Light Combination Therapy Reduces Oxidative Stress and Enhances B-Lymphocyte Proliferation through Promoting Melatonin Secretion.
Oxidative medicine and cellular longevity
Zhang Y,Wang Z,Cao J,Dong Y,Chen Y
Fri May 21 00:00:00 EDT 2021
miR-944 Suppresses EGF-Induced EMT in Colorectal Cancer Cells by Directly Targeting GATA6.
OncoTargets and therapy
Tang J,Gao W,Liu G,Sheng W,Zhou J,Dong Q,Dong M
Thu Apr 21 00:00:00 EDT 2022
miR-944 Suppresses EGF-Induced EMT in Colorectal Cancer Cells by Directly Targeting GATA6.
OncoTargets and therapy
Tang J,Gao W,Liu G,Sheng W,Zhou J,Dong Q,Dong M
Thu Apr 21 00:00:00 EDT 2022
FoxM1 promotes Wnt/β-catenin pathway activation and renal fibrosis via transcriptionally regulating multi-Wnts expressions.
Journal of cellular and molecular medicine
Xie H,Miao N,Xu D,Zhou Z,Ni J,Yin F,Wang Y,Cheng Q,Chen P,Li J,Zheng P,Zhou L,Liu J,Zhang W,Wang X,Lu L
Mon Feb 01 00:00:00 EST 2021
Type 2 Alveolar Epithelial Cells Differentiated from Human Umbilical Cord Mesenchymal Stem Cells Alleviate Mouse Pulmonary Fibrosis Through β-Catenin-Regulated Cell Apoptosis.
Stem cells and development
Liu J,Peng D,You J,Zhou O,Qiu H,Hao C,Chen H,Fu Z,Zou L
Thu Jul 01 00:00:00 EDT 2021
Inhibition of Sema4D/PlexinB1 signaling alleviates vascular dysfunction in diabetic retinopathy.
EMBO molecular medicine
Wu JH,Li YN,Chen AQ,Hong CD,Zhang CL,Wang HL,Zhou YF,Li PC,Wang Y,Mao L,Xia YP,He QW,Jin HJ,Yue ZY,Hu B
Fri Feb 07 00:00:00 EST 2020
Acute stress disrupts intestinal homeostasis via GDNF-RET.
Cell proliferation
Lin L,Feng B,Zhou R,Liu Y,Li L,Wang K,Yu Y,Liu C,Long X,Gu X,Li B,Wang X,Yang X,Cong Y,Zuo X,Li Y
Thu Oct 01 00:00:00 EDT 2020
FNDC3B 3'-UTR shortening escapes from microRNA-mediated gene repression and promotes nasopharyngeal carcinoma progression.
Cancer science
Li YQ,Chen Y,Xu YF,He QM,Yang XJ,Li YQ,Hong XH,Huang SY,Tang LL,Liu N
Mon Jun 01 00:00:00 EDT 2020
HLY78 Attenuates Neuronal Apoptosis via the LRP6/GSK3β/β-Catenin Signaling Pathway After Subarachnoid Hemorrhage in Rats.
Neuroscience bulletin
Luo X,Li L,Xu W,Cheng Y,Xie Z
Thu Oct 01 00:00:00 EDT 2020
MS-275 combined with cisplatin exerts synergistic antitumor effects in human esophageal squamous cell carcinoma cells.
Toxicology and applied pharmacology
Liu T,Guan F,Wang Y,Zhang Z,Li Y,Cui Y,Li Z,Liu H,Zhang Y,Wang Y,Ma S
Fri May 15 00:00:00 EDT 2020
Bovine Milk Exosomes Affect Proliferation and Protect Macrophages against Cisplatin-Induced Cytotoxicity.
Immunological investigations
Matic S,D'Souza DH,Wu T,Pangloli P,Dia VP
Thu Oct 01 00:00:00 EDT 2020
Bisphenol A and polychlorinated biphenyls enhance the cancer stem cell properties of human ovarian cancer cells by activating the WNT signaling pathway.
Chemosphere
Guo Y,Li B,Yan X,Shen X,Ma J,Liu S,Zhang D
Fri May 01 00:00:00 EDT 2020
BMX activates Wnt/β-catenin signaling pathway to promote cell proliferation and migration in breast cancer.
Breast cancer (Tokyo, Japan)
Li K,Pan WT,Ma YB,Xu XL,Gao Y,He YQ,Wei L,Zhang JW
Fri May 01 00:00:00 EDT 2020
Members of the miR-30 family inhibit the epithelial-to-mesenchymal transition of non-small-cell lung cancer cells by suppressing XB130 expression levels.
Oncology letters
Song K,Jiang Y,Zhao Y,Xie Y,Zhou J,Yu W,Wang Q
Thu Oct 01 00:00:00 EDT 2020
miR-224 targets BTRC and promotes cell migration and invasion in colorectal cancer.
3 Biotech
Zheng Q,Yu JJ,Li C,Li J,Wang J,Wang S
Sun Nov 01 00:00:00 EDT 2020
Endogenous GDF11 regulates odontogenic differentiation of dental pulp stem cells.
Journal of cellular and molecular medicine
Qi X,Xiao Q,Sheng R,Jiang S,Yuan Q,Liu W
Thu Oct 01 00:00:00 EDT 2020
Design and synthesis of novel Flavone-based histone deacetylase inhibitors antagonizing activation of STAT3 in breast cancer.
European journal of medicinal chemistry
Wei M,Xie M,Zhang Z,Wei Y,Zhang J,Pan H,Li B,Wang J,Song Y,Chong C,Zhao R,Wang J,Yu L,Yang G,Yang C
Sun Nov 15 00:00:00 EST 2020
Mitochondrion-Directed Nanoparticles Loaded with a Natural Compound and a microRNA for Promoting Cancer Cell Death via the Modulation of Tumor Metabolism and Mitochondrial Dynamics.
Pharmaceutics
Lo YL,Wang CS,Chen YC,Wang TY,Chang YH,Chen CJ,Yang CP
Tue Aug 11 00:00:00 EDT 2020
CTHRC1 in Ovarian Cancer Promotes M2-Like Polarization of Tumor-Associated Macrophages via Regulation of the STAT6 Signaling Pathway.
OncoTargets and therapy
Bai Y,Yin K,Su T,Ji F,Zhang S
Fri Apr 15 00:00:00 EDT 2022
Let-7b downgrades CCND1 to repress osteogenic proliferation and differentiation of MC3T3-E1 cells: An implication in osteoporosis.
The Kaohsiung journal of medical sciences
Wang LJ,Cai HQ
Thu Oct 01 00:00:00 EDT 2020
Single-cell Transcriptome Profiling reveals Dermal and Epithelial cell fate decisions during Embryonic Hair Follicle Development.
Theranostics
Ge W,Tan SJ,Wang SH,Li L,Sun XF,Shen W,Wang X
Mon May 03 00:00:00 EDT 2021
Chronic Intermittent Hypobaric Hypoxia Restores Hippocampus Function and Rescues Cognitive Impairments in Chronic Epileptic Rats via Wnt/β-catenin Signaling.
Frontiers in molecular neuroscience
Sun C,Fu J,Qu Z,Jia L,Li D,Zhen J,Wang W
Tue Feb 16 00:00:00 EST 2021
miR‑483‑3p promotes the osteogenesis of human osteoblasts by targeting Dikkopf 2 (DKK2) and the Wnt signaling pathway.
International journal of molecular medicine
Zhou B,Peng K,Wang G,Chen W,Liu P,Chen F,Kang Y
Thu Oct 01 00:00:00 EDT 2020
Heme Oxygenase-1 Suppresses Wnt Signaling Pathway in Nonalcoholic Steatohepatitis-Related Liver Fibrosis.
BioMed research international
Du J,Ren W,Zhang Q,Fu N,Han F,Cui P,Li W,Kong L,Zhao S,Wang R,Zhang Y,Yang L,Kong L,Nan Y
Fri Mar 12 00:00:00 EST 2021
Tumor Suppressor microRNA-138 Suppresses Low-Grade Glioma Development and Metastasis via Regulating IGF2BP2.
OncoTargets and therapy
Yang Y,Liu X,Cheng L,Li L,Wei Z,Wang Z,Han G,Wan X,Wang Z,Zhang J,Chen C
Wed Apr 13 00:00:00 EDT 2022
Secreted Wnt6 mediates diabetes-associated centrosome amplification via its receptor FZD4.
American journal of physiology. Cell physiology
He QJ,Wang P,Liu QQ,Wu QG,Li YF,Wang J,Lee SC
Wed Jan 01 00:00:00 EST 2020
Hyperin Controls the Development and Therapy of Gastric Cancer via Regulating Wnt/β-Catenin Signaling.
Cancer management and research
Ping MH
Mon Apr 18 00:00:00 EDT 2022
Hyperin Controls the Development and Therapy of Gastric Cancer via Regulating Wnt/β-Catenin Signaling.
Cancer management and research
Ping MH
Mon Apr 18 00:00:00 EDT 2022
BMP8A promotes survival and drug resistance via Nrf2/TRIM24 signaling pathway in clear cell renal cell carcinoma.
Cancer science
Yu YP,Cai LC,Wang XY,Cheng SY,Zhang DM,Jian WG,Wang TD,Yang JK,Yang KB,Zhang C
Fri May 01 00:00:00 EDT 2020
CHD4/NuRD complex regulates complement gene expression and correlates with CD8 T cell infiltration in human hepatocellular carcinoma.
Clinical epigenetics
Shao S,Cao H,Wang Z,Zhou D,Wu C,Wang S,Xia D,Zhang D
Tue Feb 18 00:00:00 EST 2020
CHD4/NuRD complex regulates complement gene expression and correlates with CD8 T cell infiltration in human hepatocellular carcinoma.
Clinical epigenetics
Shao S,Cao H,Wang Z,Zhou D,Wu C,Wang S,Xia D,Zhang D
Tue Feb 18 00:00:00 EST 2020
KLHL22 Regulates the EMT and Proliferation in Colorectal Cancer Cells in Part via the Wnt/β-Catenin Signaling Pathway.
Cancer management and research
Song Y,Yuan H,Wang J,Wu Y,Xiao Y,Mao S
Fri Apr 15 00:00:00 EDT 2022
GATA6 promotes epithelial-mesenchymal transition and metastasis through MUC1/β-catenin pathway in cholangiocarcinoma.
Cell death & disease
Deng X,Jiang P,Chen J,Li J,Li D,He Y,Jiang Y,Zhang Y,Xu S,Li X,Wang S,Tian F
Thu Oct 15 00:00:00 EDT 2020
GATA6 promotes epithelial-mesenchymal transition and metastasis through MUC1/β-catenin pathway in cholangiocarcinoma.
Cell death & disease
Deng X,Jiang P,Chen J,Li J,Li D,He Y,Jiang Y,Zhang Y,Xu S,Li X,Wang S,Tian F
Thu Oct 15 00:00:00 EDT 2020
GATA6 promotes epithelial-mesenchymal transition and metastasis through MUC1/β-catenin pathway in cholangiocarcinoma.
Cell death & disease
Deng X,Jiang P,Chen J,Li J,Li D,He Y,Jiang Y,Zhang Y,Xu S,Li X,Wang S,Tian F
Thu Oct 15 00:00:00 EDT 2020
The anti-migration and anti-invasion effects of Bruceine D in human triple-negative breast cancer MDA-MB-231 cells.
Experimental and therapeutic medicine
Luo C,Wang Y,Wei C,Chen Y,Ji Z
Wed Jan 01 00:00:00 EST 2020
The Wnt/β-catenin/VASP positive feedback loop drives cell proliferation and migration in breast cancer.
Oncogene
Li K,Zhang J,Tian Y,He Y,Xu X,Pan W,Gao Y,Chen F,Wei L
Sun Mar 01 00:00:00 EST 2020
The Wnt/β-catenin/VASP positive feedback loop drives cell proliferation and migration in breast cancer.
Oncogene
Li K,Zhang J,Tian Y,He Y,Xu X,Pan W,Gao Y,Chen F,Wei L
Sun Mar 01 00:00:00 EST 2020
MiR-26a-5p Serves as an Oncogenic MicroRNA in Non-Small Cell Lung Cancer by Targeting FAF1.
Cancer management and research
Ye MF,Lin D,Li WJ,Xu HP,Zhang J
Sat Apr 16 00:00:00 EDT 2022
The peptide mimicking small extracellular ring domain of CD82 inhibits epithelial-mesenchymal transition by downregulating Wnt pathway and upregulating hippo pathway.
Biochemical and biophysical research communications
He X,Huang X,Wang C,Luan M,Li Y,Ma X,Ma K
Thu Dec 10 00:00:00 EST 2020
The peptide mimicking small extracellular ring domain of CD82 inhibits epithelial-mesenchymal transition by downregulating Wnt pathway and upregulating hippo pathway.
Biochemical and biophysical research communications
He X,Huang X,Wang C,Luan M,Li Y,Ma X,Ma K
Thu Dec 10 00:00:00 EST 2020
Tissue Inhibitor of Metalloprotease-1 (TIMP-1) Regulates Adipogenesis of Adipose-derived Stem Cells (ASCs) via the Wnt Signaling Pathway in an MMP-independent Manner.
Current medical science
Wang L,Zhang CG,Jia YL,Hu L
Thu Oct 01 00:00:00 EDT 2020
Adipose-derived exosomes protect the pulmonary endothelial barrier in ventilator-induced lung injury by inhibiting the TRPV4/Ca2+ signaling pathway.
American journal of physiology. Lung cellular and molecular physiology
Yu Q,Wang D,Wen X,Tang X,Qi D,He J,Zhao Y,Deng W,Zhu T
Wed Apr 01 00:00:00 EDT 2020
LncRNA PVT1 promotes gemcitabine resistance of pancreatic cancer via activating Wnt/β-catenin and autophagy pathway through modulating the miR-619-5p/Pygo2 and miR-619-5p/ATG14 axes.
Molecular cancer
Zhou C,Yi C,Yi Y,Qin W,Yan Y,Dong X,Zhang X,Huang Y,Zhang R,Wei J,Ali DW,Michalak M,Chen XZ,Tang J
Wed Jul 29 00:00:00 EDT 2020
Nintedanib Inhibits Wnt3a-Induced Myofibroblast Activation by Suppressing the Src/β-Catenin Pathway.
Frontiers in pharmacology
Li X,Liu X,Deng R,Gao S,Yu H,Huang K,Jiang Q,Liu R,Li X,Zhang L,Zhou H,Yang C
Mon Sep 28 00:00:00 EDT 2020
Nintedanib Inhibits Wnt3a-Induced Myofibroblast Activation by Suppressing the Src/β-Catenin Pathway.
Frontiers in pharmacology
Li X,Liu X,Deng R,Gao S,Yu H,Huang K,Jiang Q,Liu R,Li X,Zhang L,Zhou H,Yang C
Mon Sep 28 00:00:00 EDT 2020
PAK6 promotes cervical cancer progression through activation of the Wnt/β-catenin signaling pathway.
Oncology letters
Yang Q,Zhao Y,Chen Y,Chang Y,Huang A,Xu T,Li G,Wu G
Tue Sep 01 00:00:00 EDT 2020
Akt inhibition-dependent downregulation of the Wnt/β-Catenin Signaling pathway contributes to antimony-induced neurotoxicity.
The Science of the total environment
Shi W,Tang Y,Zhi Y,Li Z,Yu S,Jiang J,Zhu J,Li J,Wang F,Su L,Zhao X
Thu Oct 01 00:00:00 EDT 2020
Akt inhibition-dependent downregulation of the Wnt/β-Catenin Signaling pathway contributes to antimony-induced neurotoxicity.
The Science of the total environment
Shi W,Tang Y,Zhi Y,Li Z,Yu S,Jiang J,Zhu J,Li J,Wang F,Su L,Zhao X
Thu Oct 01 00:00:00 EDT 2020
XB130, regulated by miR-203, miR-219, and miR-4782-3p, mediates the proliferation and metastasis of non-small-cell lung cancer cells.
Molecular carcinogenesis
Wang Q,Yang G,Jiang Y,Luo M,Li C,Zhao Y,Xie Y,Song K,Zhou J
Fri May 01 00:00:00 EDT 2020
The role of miR-21/RECK in the inhibition of osteosarcoma by curcumin.
Molecular and cellular probes
Zhou L,Lu Y,Liu JS,Long SZ,Liu HL,Zhang J,Zhang T
Mon Jun 01 00:00:00 EDT 2020
Musashi2 promotes EGF-induced EMT in pancreatic cancer via ZEB1-ERK/MAPK signaling.
Journal of experimental & clinical cancer research : CR
Sheng W,Shi X,Lin Y,Tang J,Jia C,Cao R,Sun J,Wang G,Zhou L,Dong M
Fri Jan 17 00:00:00 EST 2020
PYGB Promoted Tumor Progression by Regulating Wnt/β-Catenin Pathway in Gastric Cancer.
Technology in cancer research & treatment
Xia B,Zhang K,Liu C
Mon Jan 04 00:00:00 EST 2021
PYGB Promoted Tumor Progression by Regulating Wnt/β-Catenin Pathway in Gastric Cancer.
Technology in cancer research & treatment
Xia B,Zhang K,Liu C
Mon Jan 04 00:00:00 EST 2021
YTHDF1 Facilitates the Progression of Hepatocellular Carcinoma by Promoting FZD5 mRNA Translation in an m6A-Dependent Manner.
Molecular therapy. Nucleic acids
Liu X,Qin J,Gao T,Li C,He B,Pan B,Xu X,Chen X,Zeng K,Xu M,Zhu C,Pan Y,Sun H,Sun L,Xu T,Wang S
Fri Dec 04 00:00:00 EST 2020
YTHDF1 Facilitates the Progression of Hepatocellular Carcinoma by Promoting FZD5 mRNA Translation in an m6A-Dependent Manner.
Molecular therapy. Nucleic acids
Liu X,Qin J,Gao T,Li C,He B,Pan B,Xu X,Chen X,Zeng K,Xu M,Zhu C,Pan Y,Sun H,Sun L,Xu T,Wang S
Fri Dec 04 00:00:00 EST 2020
Caveolin-1 upregulates Fut8 expression by activating the Wnt/β-catenin pathway to enhance HCC cell proliferative and invasive ability.
Cell biology international
Zhang C,Wu Q,Huang H,Chen X,Huang T,Li W,Zhang J,Liu Y
Sun Nov 01 00:00:00 EDT 2020
Long noncoding RNA CA7-4 promotes autophagy and apoptosis via sponging MIR877-3P and MIR5680 in high glucose-induced vascular endothelial cells.
Autophagy
Zhao X,Su L,He X,Zhao B,Miao J
Wed Jan 01 00:00:00 EST 2020
HLY78 protects blood-brain barrier integrity through Wnt/β-catenin signaling pathway following subarachnoid hemorrhage in rats.
Brain research bulletin
Luo X,Li L,Zheng W,Gu L,Zhang X,Li Y,Xie Z,Cheng Y
Tue Sep 01 00:00:00 EDT 2020
A Cdh1-FoxM1-Apc axis controls muscle development and regeneration.
Cell death & disease
Chen Z,Li L,Xu S,Liu Z,Zhou C,Li Z,Liu Y,Wu W,Huang Y,Kuang M,Fan S,Li H,Li X,Song G,Wu WS,Chen J,Hou Y
Mon Mar 09 00:00:00 EDT 2020
A Cdh1-FoxM1-Apc axis controls muscle development and regeneration.
Cell death & disease
Chen Z,Li L,Xu S,Liu Z,Zhou C,Li Z,Liu Y,Wu W,Huang Y,Kuang M,Fan S,Li H,Li X,Song G,Wu WS,Chen J,Hou Y
Mon Mar 09 00:00:00 EDT 2020
1,25-dihydroxyvitamin D3 signaling-induced decreases in IRX4 inhibits NANOG-mediated cancer stem-like properties and gefitinib resistance in NSCLC cells.
Cell death & disease
Jia Z,Zhang Y,Yan A,Wang M,Han Q,Wang K,Wang J,Qiao C,Pan Z,Chen C,Hu D,Ding X
Thu Aug 20 00:00:00 EDT 2020
Cancer-Testis Gene Expression in Hepatocellular Carcinoma: Identification of Prognostic Markers and Potential Targets for Immunotherapy.
Technology in cancer research & treatment
Zhang YP,Bao ZW,Wu JB,Chen YH,Chen JR,Xie HY,Zhou L,Wu J,Zheng SS
Mon Nov 29 00:00:00 EST 2021
Liraglutide regulates proliferation, differentiation, and apoptosis of preosteoblasts through a signaling network of Notch/Wnt/Hedgehog signaling pathways.
European review for medical and pharmacological sciences
Hou HW,Xue P,Wang Y,Li YK
Tue Dec 01 00:00:00 EST 2020
Liraglutide regulates proliferation, differentiation, and apoptosis of preosteoblasts through a signaling network of Notch/Wnt/Hedgehog signaling pathways.
European review for medical and pharmacological sciences
Hou HW,Xue P,Wang Y,Li YK
Tue Dec 01 00:00:00 EST 2020
BAIAP2L2 promotes the progression of gastric cancer via AKT/mTOR and Wnt3a/β-catenin signaling pathways.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Liu J,Shangguan Y,Sun J,Cong W,Xie Y
Tue Sep 01 00:00:00 EDT 2020
TMED3 Promotes Proliferation and Migration in Breast Cancer Cells by Activating Wnt/β-Catenin Signaling.
OncoTargets and therapy
Zhang X,Luo Y,Li Q
Fri Apr 15 00:00:00 EDT 2022
TMED3 Promotes Proliferation and Migration in Breast Cancer Cells by Activating Wnt/β-Catenin Signaling.
OncoTargets and therapy
Zhang X,Luo Y,Li Q
Fri Apr 15 00:00:00 EDT 2022
Lgr4 Deletion Delays the Hair Cycle and Inhibits the Activation of Hair Follicle Stem Cells.
The Journal of investigative dermatology
Ren X,Xia W,Xu P,Shen H,Dai X,Liu M,Shi Y,Ye X,Dang Y
Tue Sep 01 00:00:00 EDT 2020
Strontium modulates osteogenic activity of bone cement composed of bioactive borosilicate glass particles by activating Wnt/β-catenin signaling pathway.
Bioactive materials
Cui X,Zhang Y,Wang J,Huang C,Wang Y,Yang H,Liu W,Wang T,Wang D,Wang G,Ruan C,Chen D,Lu WW,Huang W,Rahaman MN,Pan H
Mon Jun 01 00:00:00 EDT 2020
MiR-377-3p suppresses colorectal cancer through negative regulation on Wnt/β-catenin signaling by targeting XIAP and ZEB2.
Pharmacological research
Huang L,Liu Z,Hu J,Luo Z,Zhang C,Wang L,Wang Z
Mon Jun 01 00:00:00 EDT 2020
Down-regulation of exosomal miR-200c derived from keratinocytes in vitiligo lesions suppresses melanogenesis.
Journal of cellular and molecular medicine
Zhao C,Wang D,Wang X,Mao Y,Xu Z,Sun Y,Mei X,Song J,Shi W
Thu Oct 01 00:00:00 EDT 2020
Although c‑MYC contributes to tamoxifen resistance, it improves cisplatin sensitivity in ER‑positive breast cancer.
International journal of oncology
Chen R,Guo S,Yang C,Sun L,Zong B,Li K,Liu L,Tu G,Liu M,Liu S
Wed Apr 01 00:00:00 EDT 2020
Crosstalk between androgen and Wnt/β-catenin leads to changes of wool density in FGF5-knockout sheep.
Cell death & disease
Zhang R,Li Y,Jia K,Xu X,Li Y,Zhao Y,Zhang X,Zhang J,Liu G,Deng S,Lian Z
Fri May 29 00:00:00 EDT 2020
A novel Sprouty4-ERK1/2-Wnt/β-catenin regulatory loop in marrow stromal progenitor cells controls osteogenic and adipogenic differentiation.
Metabolism: clinical and experimental
Tian L,Xiao H,Li M,Wu X,Xie Y,Zhou J,Zhang X,Wang B
Wed Apr 01 00:00:00 EDT 2020
Involvement of the ERK/HIF-1α/EMT Pathway in XCL1-Induced Migration of MDA-MB-231 and SK-BR-3 Breast Cancer Cells.
International journal of molecular sciences
Do HTT,Cho J
Wed Dec 23 00:00:00 EST 2020
Involvement of the ERK/HIF-1α/EMT Pathway in XCL1-Induced Migration of MDA-MB-231 and SK-BR-3 Breast Cancer Cells.
International journal of molecular sciences
Do HTT,Cho J
Wed Dec 23 00:00:00 EST 2020
Norepinephrine-CREB1-miR-373 axis promotes progression of colon cancer.
Molecular oncology
Han J,Jiang Q,Ma R,Zhang H,Tong D,Tang K,Wang X,Ni L,Miao J,Duan B,Yang Y,Chen Y,Wu F,Han J,Wang M,Hou N,Huang C
Fri May 01 00:00:00 EDT 2020
Inhibition of GSK3 Represses the Expression of Retinoic Acid Synthetic Enzyme ALDH1A2 via Wnt/β-Catenin Signaling in WiT49 Cells.
Frontiers in cell and developmental biology
Li Y,Gong H,Ding J,Zhao F,Du J,Wan J,Zhang J,Liu S,Li J,Wang L,Zhou B
Mon Sep 28 00:00:00 EDT 2020
Inhibition of GSK3 Represses the Expression of Retinoic Acid Synthetic Enzyme ALDH1A2 via Wnt/β-Catenin Signaling in WiT49 Cells.
Frontiers in cell and developmental biology
Li Y,Gong H,Ding J,Zhao F,Du J,Wan J,Zhang J,Liu S,Li J,Wang L,Zhou B
Mon Sep 28 00:00:00 EDT 2020
Inhibition of GSK3 Represses the Expression of Retinoic Acid Synthetic Enzyme ALDH1A2 via Wnt/β-Catenin Signaling in WiT49 Cells.
Frontiers in cell and developmental biology
Li Y,Gong H,Ding J,Zhao F,Du J,Wan J,Zhang J,Liu S,Li J,Wang L,Zhou B
Mon Sep 28 00:00:00 EDT 2020
Lysophosphatidic acid receptor 4 regulates osteogenic and adipogenic differentiation of progenitor cells via inactivation of RhoA/ROCK1/β-catenin signaling.
Stem cells (Dayton, Ohio)
Xie Y,Wang X,Wu X,Tian L,Zhou J,Li X,Wang B
Sun Mar 01 00:00:00 EST 2020
Transcription co-activator P300 activates Elk1-aPKC-ι signaling mediated epithelial-to-mesenchymal transition and malignancy in hepatocellular carcinoma.
Oncogenesis
Ma C,Huang S,Xu L,Tian L,Yang Y,Wang J
Fri Mar 06 00:00:00 EST 2020
Diethyldithiocarbamate-copper nanocomplex reinforces disulfiram chemotherapeutic efficacy through light-triggered nuclear targeting.
Theranostics
Ren L,Feng W,Shao J,Ma J,Xu M,Zhu BZ,Zheng N,Liu S
Wed Apr 28 00:00:00 EDT 2021
Anthocyanins from Aronia melanocarpa Induce Apoptosis in Caco-2 Cells through Wnt/β-Catenin Signaling Pathway.
Chemistry & biodiversity
Wei J,Yu W,Hao R,Fan J,Gao J
Sun Nov 01 00:00:00 EDT 2020
AGR3 promotes the stemness of colorectal cancer via modulating Wnt/β-catenin signalling.
Cellular signalling
Chi J,Zhang H,Hu J,Song Y,Li J,Wang L,Wang Z
Wed Jan 01 00:00:00 EST 2020
Aberrant DNA hypermethylation-silenced LINC00886 gene accelerates malignant progression of laryngeal carcinoma.
Pathology, research and practice
Lan L,Cao H,Chi W,Meng W,Zhao L,Cui W,Wang B
Wed Apr 01 00:00:00 EDT 2020
TAp63α targeting of Lgr5 mediates colorectal cancer stem cell properties and sulforaphane inhibition.
Oncogenesis
Chen Y,Wang MH,Zhu JY,Xie CF,Li XT,Wu JS,Geng SS,Han HY,Zhong CY
Sat Oct 10 00:00:00 EDT 2020
Melatonin protects the mouse testis against heat-induced damage.
Molecular human reproduction
Zhang P,Zheng Y,Lv Y,Li F,Su L,Qin Y,Zeng W
Sat Feb 29 00:00:00 EST 2020
ADNP promotes neural differentiation by modulating Wnt/β-catenin signaling.
Nature communications
Sun X,Peng X,Cao Y,Zhou Y,Sun Y
Fri Jun 12 00:00:00 EDT 2020
SMYD2 suppresses APC2 expression to activate the Wnt/β-catenin pathway and promotes epithelial-mesenchymal transition in colorectal cancer.
American journal of cancer research
Meng F,Liu X,Lin C,Xu L,Liu J,Zhang P,Zhang X,Song J,Yan Y,Ren Z,Zhang Y
Mon Sep 28 00:00:00 EDT 2020
Therapeutic effect of GLP-1 engineered strain on mice model of Alzheimer's disease and Parkinson's disease.
AMB Express
Fang X,Zhou X,Miao Y,Han Y,Wei J,Chen T
Fri Apr 24 00:00:00 EDT 2020
MicroRNA-1236-3p inhibits human osteosarcoma growth.
Oncology letters
Li J,Chen J,Hu Z,Xu W
Tue Dec 01 00:00:00 EST 2020
MicroRNA-1236-3p inhibits human osteosarcoma growth.
Oncology letters
Li J,Chen J,Hu Z,Xu W
Tue Dec 01 00:00:00 EST 2020
Combination of Inositol Hexaphosphate and Inositol Inhibits Liver Metastasis of Colorectal Cancer in Mice Through the Wnt/β-Catenin Pathway.
OncoTargets and therapy
Liu X,Liu C,Chen C,Sun W,Ci Y,Li Q,Song Y
Thu Apr 14 00:00:00 EDT 2022
Linc00261 inhibits metastasis and the WNT signaling pathway of pancreatic cancer by regulating a miR‑552‑5p/FOXO3 axis.
Oncology reports
Chen T,Lei S,Zeng Z,Zhang J,Xue Y,Sun Y,Lan J,Xu S,Mao D,Guo B
Sun Mar 01 00:00:00 EST 2020
β-catenin stimulates Tcf7l1 degradation through recruitment of casein kinase 2 in mouse embryonic stem cells.
Biochemical and biophysical research communications
Zhang Y,Zhu Z,Ding H,Wan S,Zhang X,Li Y,Ji J,Wang X,Zhang M,Ye SD
Thu Apr 02 00:00:00 EDT 2020
β-catenin stimulates Tcf7l1 degradation through recruitment of casein kinase 2 in mouse embryonic stem cells.
Biochemical and biophysical research communications
Zhang Y,Zhu Z,Ding H,Wan S,Zhang X,Li Y,Ji J,Wang X,Zhang M,Ye SD
Thu Apr 02 00:00:00 EDT 2020
ALDH1A1 maintains the cancer stem-like cells properties of esophageal squamous cell carcinoma by activating the AKT signal pathway and interacting with β-catenin.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang W,He S,Zhang R,Peng J,Guo D,Zhang J,Xiang B,Li L
Fri May 01 00:00:00 EDT 2020
Musashi2 promotes the progression of pancreatic cancer through a novel ISYNA1-p21/ZEB-1 pathway.
Journal of cellular and molecular medicine
Zhou L,Sheng W,Jia C,Shi X,Cao R,Wang G,Lin Y,Zhu F,Dong Q,Dong M
Tue Sep 01 00:00:00 EDT 2020
Inhibition of p66Shc Oxidative Signaling via CA-Induced Upregulation of miR-203a-3p Alleviates Liver Fibrosis Progression.
Molecular therapy. Nucleic acids
Wang Z,Zhao Y,Zhao H,Zhou J,Feng D,Tang F,Li Y,Lv L,Chen Z,Ma X,Tian X,Yao J
Fri Sep 04 00:00:00 EDT 2020
Silencing of the lncRNA TUG1 attenuates the epithelial-mesenchymal transition of renal tubular epithelial cells by sponging miR-141-3p via regulating β-catenin.
American journal of physiology. Renal physiology
Zhang B,Zhao C,Hou L,Wu Y
Tue Dec 01 00:00:00 EST 2020
N-glycosylation of uterine endometrium determines its receptivity.
Journal of cellular physiology
Yu M,Qin H,Wang H,Liu J,Liu S,Yan Q
Sat Feb 01 00:00:00 EST 2020
TBC1D20 Is Essential for Mouse Blood-Testis Barrier Integrity Through Maintaining the Epithelial Phenotype and Modulating the Maturation of Sertoli Cells.
Reproductive sciences (Thousand Oaks, Calif.)
Cui L,Gu Y,Liu S,Li M,Ye J,Zhang F,Luo X,Chang WL,Gui Y
Wed Jul 01 00:00:00 EDT 2020
Identification of a subpopulation of long-term tumor-initiating cells in colon cancer.
Bioscience reports
Peng L,Xiong Y,Wang R,Xiang L,Zhou H,Gu H
Fri Aug 28 00:00:00 EDT 2020
Circular RNA circREPS2 Acts as a Sponge of miR-558 to Suppress Gastric Cancer Progression by Regulating RUNX3/β-catenin Signaling.
Molecular therapy. Nucleic acids
Guo X,Dai X,Liu J,Cheng A,Qin C,Wang Z
Fri Sep 04 00:00:00 EDT 2020
A Noncoding Regulatory RNAs Network Driven by Circ-CDYL Acts Specifically in the Early Stages Hepatocellular Carcinoma.
Hepatology (Baltimore, Md.)
Wei Y,Chen X,Liang C,Ling Y,Yang X,Ye X,Zhang H,Yang P,Cui X,Ren Y,Xin X,Li H,Wang R,Wang W,Jiang F,Liu S,Ding J,Zhang B,Li L,Wang H
Wed Jan 01 00:00:00 EST 2020
Integrative Analysis of Methylome and Transcriptome Reveals the Regulatory Mechanisms of Hair Follicle Morphogenesis in Cashmere Goat.
Cells
Wang S,Li F,Liu J,Zhang Y,Zheng Y,Ge W,Qu L,Wang X
Tue Apr 14 00:00:00 EDT 2020
Upregulation of miR-92a-2-5p potentially contribute to anorectal malformations by inhibiting proliferation and enhancing apoptosis via PRKCA/β-catenin.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Long CY,Xiao YX,Li SY,Tang XB,Yuan ZW,Bai YZ
Wed Jul 01 00:00:00 EDT 2020
Liraglutide-loaded PLGA/gelatin electrospun nanofibrous mats promote angiogenesis to accelerate diabetic wound healing via the modulation of miR-29b-3p.
Biomaterials science
Yu M,Huang J,Zhu T,Lu J,Liu J,Li X,Yan X,Liu F
Fri Aug 07 00:00:00 EDT 2020
MicroRNA‑137 suppresses the proliferation, migration and invasion of cholangiocarcinoma cells by targeting WNT2B.
International journal of molecular medicine
Chen T,Lei S,Zeng Z,Pan S,Zhang J,Xue Y,Sun Y,Lan J,Xu S,Mao D,Guo B
Sun Mar 01 00:00:00 EST 2020
Circulating Exosomes From Lipopolysaccharide-Induced Ards Mice Trigger Endoplasmic Reticulum Stress in Lung Tissue.
Shock (Augusta, Ga.)
Tang X,Yu Q,Wen X,Qi D,Peng J,He J,Deng W,Zhu T,Zhao Y,Wang D
Wed Jul 01 00:00:00 EDT 2020
FAM172A inhibits EMT in pancreatic cancer via ERK-MAPK signaling.
Biology open
Chen Y,Liu P,Shen D,Liu H,Xu L,Wang J,Shen D,Sun H,Wu H
Fri Feb 07 00:00:00 EST 2020
ADAM10 promotes cell growth, migration, and invasion in osteosarcoma via regulating E-cadherin/β-catenin signaling pathway and is regulated by miR-122-5p.
Cancer cell international
Yuan Q,Yu H,Chen J,Song X,Sun L
Wed Apr 13 00:00:00 EDT 2022
miR-370-3p Alleviates Ulcerative Colitis-Related Colorectal Cancer in Mice Through Inhibiting the Inflammatory Response and Epithelial-Mesenchymal Transition.
Drug design, development and therapy
Lin L,Wang D,Qu S,Zhao H,Lin Y
Thu Feb 04 00:00:00 EST 2021
Novel HSP90-PI3K Dual Inhibitor Suppresses Melanoma Cell Proliferation by Interfering with HSP90-EGFR Interaction and Downstream Signaling Pathways.
International journal of molecular sciences
Zhao Q,Zhu HP,Xie X,Mao Q,Liu YQ,He XH,Peng C,Jiang QL,Huang W
Sat Mar 07 00:00:00 EST 2020
The dynamic expression of canonical Wnt/β-catenin signalling pathway in the pathologic process of experimental autoimmune neuritis.
The International journal of neuroscience
Liu Y,Liu S,Pan S,Gong Q,Yao J,Lu Z
Sun Nov 01 00:00:00 EDT 2020
lncRNA SLC7A11-AS1 Promotes Chemoresistance by Blocking SCFβ-TRCP-Mediated Degradation of NRF2 in Pancreatic Cancer.
Molecular therapy. Nucleic acids
Yang Q,Li K,Huang X,Zhao C,Mei Y,Li X,Jiao L,Yang H
Fri Mar 06 00:00:00 EST 2020
Baohuoside i suppresses breast cancer metastasis by downregulating the tumor-associated macrophages/C-X-C motif chemokine ligand 1 pathway.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Wang S,Wang N,Huang X,Yang B,Zheng Y,Zhang J,Wang X,Lin Y,Wang Z
Sun Nov 01 00:00:00 EDT 2020
OIP5-AS1 contributes to tumorigenesis in hepatocellular carcinoma by miR-300/YY1-activated WNT pathway.
Cancer cell international
Wang Y,Dou L,Qin Y,Yang H,Yan P
Sat Apr 16 00:00:00 EDT 2022
Sini San ameliorates duodenal mucosal barrier injury and low‑grade inflammation via the CRF pathway in a rat model of functional dyspepsia.
International journal of molecular medicine
Zhu C,Zhao L,Zhao J,Zhang S
Wed Jan 01 00:00:00 EST 2020
CELSR1 Promotes Neuroprotection in Cerebral Ischemic Injury Mainly Through the Wnt/PKC Signaling Pathway.
International journal of molecular sciences
Wang LH,Zhang GL,Liu XY,Peng A,Ren HY,Huang SH,Liu T,Wang XJ
Thu Feb 13 00:00:00 EST 2020
Mitochondrial Dynamics Is Critical for the Full Pluripotency and Embryonic Developmental Potential of Pluripotent Stem Cells.
Cell metabolism
Zhong X,Cui P,Cai Y,Wang L,He X,Long P,Lu K,Yan R,Zhang Y,Pan X,Zhao X,Li W,Zhang H,Zhou Q,Gao P
Tue Apr 02 00:00:00 EDT 2019
Mitochondrial Dynamics Is Critical for the Full Pluripotency and Embryonic Developmental Potential of Pluripotent Stem Cells.
Cell metabolism
Zhong X,Cui P,Cai Y,Wang L,He X,Long P,Lu K,Yan R,Zhang Y,Pan X,Zhao X,Li W,Zhang H,Zhou Q,Gao P
Tue Apr 02 00:00:00 EDT 2019
Mitochondrial Dynamics Is Critical for the Full Pluripotency and Embryonic Developmental Potential of Pluripotent Stem Cells.
Cell metabolism
Zhong X,Cui P,Cai Y,Wang L,He X,Long P,Lu K,Yan R,Zhang Y,Pan X,Zhao X,Li W,Zhang H,Zhou Q,Gao P
Tue Apr 02 00:00:00 EDT 2019
Mitochondrial Dynamics Is Critical for the Full Pluripotency and Embryonic Developmental Potential of Pluripotent Stem Cells.
Cell metabolism
Zhong X,Cui P,Cai Y,Wang L,He X,Long P,Lu K,Yan R,Zhang Y,Pan X,Zhao X,Li W,Zhang H,Zhou Q,Gao P
Tue Apr 02 00:00:00 EDT 2019
Mitochondrial Dynamics Is Critical for the Full Pluripotency and Embryonic Developmental Potential of Pluripotent Stem Cells.
Cell metabolism
Zhong X,Cui P,Cai Y,Wang L,He X,Long P,Lu K,Yan R,Zhang Y,Pan X,Zhao X,Li W,Zhang H,Zhou Q,Gao P
Tue Apr 02 00:00:00 EDT 2019
Mitochondrial Dynamics Is Critical for the Full Pluripotency and Embryonic Developmental Potential of Pluripotent Stem Cells.
Cell metabolism
Zhong X,Cui P,Cai Y,Wang L,He X,Long P,Lu K,Yan R,Zhang Y,Pan X,Zhao X,Li W,Zhang H,Zhou Q,Gao P
Tue Apr 02 00:00:00 EDT 2019
Mitochondrial Dynamics Is Critical for the Full Pluripotency and Embryonic Developmental Potential of Pluripotent Stem Cells.
Cell metabolism
Zhong X,Cui P,Cai Y,Wang L,He X,Long P,Lu K,Yan R,Zhang Y,Pan X,Zhao X,Li W,Zhang H,Zhou Q,Gao P
Tue Apr 02 00:00:00 EDT 2019
Cellular Internalization-Induced Aggregation of Porous Silicon Nanoparticles for Ultrasound Imaging and Protein-Mediated Protection of Stem Cells.
Small (Weinheim an der Bergstrasse, Germany)
Qi S,Zhang P,Ma M,Yao M,Wu J,Mäkilä E,Salonen J,Ruskoaho H,Xu Y,Santos HA,Zhang H
Tue Jan 01 00:00:00 EST 2019
Cellular Internalization-Induced Aggregation of Porous Silicon Nanoparticles for Ultrasound Imaging and Protein-Mediated Protection of Stem Cells.
Small (Weinheim an der Bergstrasse, Germany)
Qi S,Zhang P,Ma M,Yao M,Wu J,Mäkilä E,Salonen J,Ruskoaho H,Xu Y,Santos HA,Zhang H
Tue Jan 01 00:00:00 EST 2019
Tumor-suppressive functions of 4-MU on breast cancer cells of different ER status: Regulation of hyaluronan/HAS2/CD44 and specific matrix effectors.
Matrix biology : journal of the International Society for Matrix Biology
Karalis TT,Heldin P,Vynios DH,Neill T,Buraschi S,Iozzo RV,Karamanos NK,Skandalis SS
Wed May 01 00:00:00 EDT 2019
Tumor-suppressive functions of 4-MU on breast cancer cells of different ER status: Regulation of hyaluronan/HAS2/CD44 and specific matrix effectors.
Matrix biology : journal of the International Society for Matrix Biology
Karalis TT,Heldin P,Vynios DH,Neill T,Buraschi S,Iozzo RV,Karamanos NK,Skandalis SS
Wed May 01 00:00:00 EDT 2019
Metformin Improves Fertility in Obese Males by Alleviating Oxidative Stress-Induced Blood-Testis Barrier Damage.
Oxidative medicine and cellular longevity
Ye J,Luo D,Xu X,Sun M,Su X,Tian Z,Zhang M,Yu C,Guan Q
Fri Mar 20 00:00:00 EDT 2020
miR-375-3p/YWHAZ/β-catenin axis regulates migration, invasion, EMT in gastric cancer cells.
Clinical and experimental pharmacology & physiology
Guo F,Gao Y,Sui G,Jiao D,Sun L,Fu Q,Jin C
Fri Feb 01 00:00:00 EST 2019
miR-375-3p/YWHAZ/β-catenin axis regulates migration, invasion, EMT in gastric cancer cells.
Clinical and experimental pharmacology & physiology
Guo F,Gao Y,Sui G,Jiao D,Sun L,Fu Q,Jin C
Fri Feb 01 00:00:00 EST 2019
miR-375-3p/YWHAZ/β-catenin axis regulates migration, invasion, EMT in gastric cancer cells.
Clinical and experimental pharmacology & physiology
Guo F,Gao Y,Sui G,Jiao D,Sun L,Fu Q,Jin C
Fri Feb 01 00:00:00 EST 2019
miR-375-3p/YWHAZ/β-catenin axis regulates migration, invasion, EMT in gastric cancer cells.
Clinical and experimental pharmacology & physiology
Guo F,Gao Y,Sui G,Jiao D,Sun L,Fu Q,Jin C
Fri Feb 01 00:00:00 EST 2019
Expression of ARHGAP10 correlates with prognosis of prostate cancer.
International journal of clinical and experimental pathology
Gong H,Chen X,Jin Y,Lu J,Cai Y,Wei O,Zhao J,Zhang W,Wen X,Wang Y,Chen W
Fri Jan 17 00:00:00 EST 2020
Acetylation of PHF5A Modulates Stress Responses and Colorectal Carcinogenesis through Alternative Splicing-Mediated Upregulation of KDM3A.
Molecular cell
Wang Z,Yang X,Liu C,Li X,Zhang B,Wang B,Zhang Y,Song C,Zhang T,Liu M,Liu B,Ren M,Jiang H,Zou J,Liu X,Zhang H,Zhu WG,Yin Y,Zhang Z,Gu W,Luo J
Thu Jun 20 00:00:00 EDT 2019
A panel of biomarkers for skin squamous cell carcinoma: various functional entities and differential responses to resveratrol.
International journal of clinical and experimental pathology
Sun Y,Li A,Liu X,Wang Q,Bai Y,Liu Z,Huang L,Wu M,Li H,Miao J,Liu J
Fri Jan 17 00:00:00 EST 2020
Glycogen phosphorylase B promotes ovarian cancer progression via Wnt/β-catenin signaling and is regulated by miR-133a-3p.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhou Y,Jin Z,Wang C
Sun Dec 01 00:00:00 EST 2019
Secalonic Acid-F, a Novel Mycotoxin, Represses the Progression of Hepatocellular Carcinoma via MARCH1 Regulation of the PI3K/AKT/β-catenin Signaling Pathway.
Molecules (Basel, Switzerland)
Xie L,Li M,Liu D,Wang X,Wang P,Dai H,Yang W,Liu W,Hu X,Zhao M
Tue Jan 22 00:00:00 EST 2019
Ivermectin inhibits canine mammary tumor growth by regulating cell cycle progression and WNT signaling.
BMC veterinary research
Diao H,Cheng N,Zhao Y,Xu H,Dong H,Thamm DH,Zhang D,Lin D
Fri Aug 02 00:00:00 EDT 2019
MiR-26b suppresses hepatocellular carcinoma development by negatively regulating ZNRD1 and Wnt/β-catenin signaling.
Cancer medicine
Hu X,Wang R,Ren Z,Liu X,Gu J,Cui G,Li Q
Sun Dec 01 00:00:00 EST 2019
Cinobufotalin powerfully reversed EBV-miR-BART22-induced cisplatin resistance via stimulating MAP2K4 to antagonize non-muscle myosin heavy chain IIA/glycogen synthase 3β/β-catenin signaling pathway.
EBioMedicine
Liu Y,Jiang Q,Liu X,Lin X,Tang Z,Liu C,Zhou J,Zhao M,Li X,Cheng Z,Li L,Xie Y,Liu Z,Fang W
Tue Oct 01 00:00:00 EDT 2019
Cinobufotalin powerfully reversed EBV-miR-BART22-induced cisplatin resistance via stimulating MAP2K4 to antagonize non-muscle myosin heavy chain IIA/glycogen synthase 3β/β-catenin signaling pathway.
EBioMedicine
Liu Y,Jiang Q,Liu X,Lin X,Tang Z,Liu C,Zhou J,Zhao M,Li X,Cheng Z,Li L,Xie Y,Liu Z,Fang W
Tue Oct 01 00:00:00 EDT 2019
ERα and Wnt/β‑catenin signaling pathways are involved in angelicin‑dependent promotion of osteogenesis.
Molecular medicine reports
Ge L,Cui Y,Liu B,Yin X,Pang J,Han J
Wed May 01 00:00:00 EDT 2019
TRIM28 protects CARM1 from proteasome-mediated degradation to prevent colorectal cancer metastasis.
Science bulletin
Cui J,Hu J,Ye Z,Fan Y,Li Y,Wang G,Wang L,Wang Z
Tue Jul 30 00:00:00 EDT 2019
Rapamycin attenuates articular cartilage degeneration by inhibiting β-catenin in a murine model of osteoarthritis.
Connective tissue research
Ma L,Liu Y,Zhao X,Li P,Jin Q
Sun Sep 01 00:00:00 EDT 2019
Targeting glutaminase 1 attenuates stemness properties in hepatocellular carcinoma by increasing reactive oxygen species and suppressing Wnt/beta-catenin pathway.
EBioMedicine
Li B,Cao Y,Meng G,Qian L,Xu T,Yan C,Luo O,Wang S,Wei J,Ding Y,Yu D
Tue Jan 01 00:00:00 EST 2019
Targeting glutaminase 1 attenuates stemness properties in hepatocellular carcinoma by increasing reactive oxygen species and suppressing Wnt/beta-catenin pathway.
EBioMedicine
Li B,Cao Y,Meng G,Qian L,Xu T,Yan C,Luo O,Wang S,Wei J,Ding Y,Yu D
Tue Jan 01 00:00:00 EST 2019
MIR17HG-miR-18a/19a axis, regulated by interferon regulatory factor-1, promotes gastric cancer metastasis via Wnt/β-catenin signalling.
Cell death & disease
Yuan J,Tan L,Yin Z,Zhu W,Tao K,Wang G,Shi W,Gao J
Tue Jun 11 00:00:00 EDT 2019
MIR17HG-miR-18a/19a axis, regulated by interferon regulatory factor-1, promotes gastric cancer metastasis via Wnt/β-catenin signalling.
Cell death & disease
Yuan J,Tan L,Yin Z,Zhu W,Tao K,Wang G,Shi W,Gao J
Tue Jun 11 00:00:00 EDT 2019
Astaxanthin protects against renal fibrosis through inhibiting myofibroblast activation and promoting CD8+ T cell recruitment.
Biochimica et biophysica acta. General subjects
Diao W,Chen W,Cao W,Yuan H,Ji H,Wang T,Chen W,Zhu X,Zhou H,Guo H,Zhao X
Sun Sep 01 00:00:00 EDT 2019
Baicalin induced colon cancer cells apoptosis through miR-217/DKK1-mediated inhibition of Wnt signaling pathway.
Molecular biology reports
Jia Y,Chen L,Guo S,Li Y
Mon Apr 01 00:00:00 EDT 2019
TROAP regulates prostate cancer progression via the WNT3/survivin signalling pathways.
Oncology reports
Ye J,Chu C,Chen M,Shi Z,Gan S,Qu F,Pan X,Yang Q,Tian Y,Wang L,Yang W,Cui X
Fri Feb 01 00:00:00 EST 2019
Chronic CagA-positive Helicobacter pylori infection with MNNG stimulation synergistically induces mesenchymal and cancer stem cell-like properties in gastric mucosal epithelial cells.
Journal of cellular biochemistry
Lin L,Wei H,Yi J,Xie B,Chen J,Zhou C,Wang L,Yang Y
Tue Oct 01 00:00:00 EDT 2019
Histone demethylase KDM5A is transactivated by the transcription factor C/EBPβ and promotes preadipocyte differentiation by inhibiting Wnt/β-catenin signaling.
The Journal of biological chemistry
Guo L,Guo YY,Li BY,Peng WQ,Tang QQ
Fri Jun 14 00:00:00 EDT 2019
ADAR1 may be involved in the proliferation of acute myeloid leukemia cells via regulation of the Wnt pathway.
Cancer management and research
Xiao H,Cheng Q,Wu X,Tang Y,Liu J,Li X
Sun Apr 10 00:00:00 EDT 2022
Mycoplasma hyorhinis infection promotes gastric cancer cell motility via β-catenin signaling.
Cancer medicine
Liu X,Rong Z,Shou C
Sun Sep 01 00:00:00 EDT 2019
Mycoplasma hyorhinis infection promotes gastric cancer cell motility via β-catenin signaling.
Cancer medicine
Liu X,Rong Z,Shou C
Sun Sep 01 00:00:00 EDT 2019
A mechanism of regucalcin knock-down in the promotion of proliferation and movement of human cervical cancer HeLa cells.
Translational cancer research
Li X,Huang Y,Wang P,Song W,Yao Q,Hu Q,Zhou S
Mon Apr 01 00:00:00 EDT 2019
Design, synthesis and molecular mechanisms of novel dual inhibitors of heat shock protein 90/phosphoinositide 3-kinase alpha (Hsp90/PI3Kα) against cutaneous melanoma.
Journal of enzyme inhibition and medicinal chemistry
Qin F,Wang Y,Jiang X,Wang Y,Zhang N,Wen X,Wang L,Jiang Q,He G
Sun Dec 01 00:00:00 EST 2019
Recombinant porcine NK-lysin inhibits the invasion of hepatocellular carcinoma cells in vitro.
International journal of biological macromolecules
Khan A,Fan K,Sun N,Yin W,Sun Y,Sun P,Jahejo AR,Li H
Fri Nov 01 00:00:00 EDT 2019
Effects of SIRT1 silencing on viability, invasion and metastasis of human glioma cell lines.
Oncology letters
Li Y,Chen X,Cui Y,Wei Q,Chen S,Wang X
Mon Apr 01 00:00:00 EDT 2019
MARCH1 encourages tumour progression of hepatocellular carcinoma via regulation of PI3K-AKT-β-catenin pathways.
Journal of cellular and molecular medicine
Xie L,Dai H,Li M,Yang W,Yu G,Wang X,Wang P,Liu W,Hu X,Zhao M
Wed May 01 00:00:00 EDT 2019
A cellular model for Wilson's disease using patient-derived induced pluripotent stem cells revealed aberrant β-catenin pathway during osteogenesis.
Biochemical and biophysical research communications
Liu J,Cui Y,Shi L,Luan J,Zhou X,Han J
Tue May 28 00:00:00 EDT 2019
A cellular model for Wilson's disease using patient-derived induced pluripotent stem cells revealed aberrant β-catenin pathway during osteogenesis.
Biochemical and biophysical research communications
Liu J,Cui Y,Shi L,Luan J,Zhou X,Han J
Tue May 28 00:00:00 EDT 2019
Inhibition of fatty acid synthesis arrests colorectal neoplasm growth and metastasis: Anti-cancer therapeutical effects of natural cyclopeptide RA-XII.
Biochemical and biophysical research communications
Wang Y,Guo D,He J,Song L,Chen H,Zhang Z,Tan N
Tue May 14 00:00:00 EDT 2019
A novel oncogene TRIM63 promotes cell proliferation and migration via activating Wnt/β-catenin signaling pathway in breast cancer.
Pathology, research and practice
Li K,Pan W,Ma Y,Xu X,Gao Y,He Y,Wei L,Zhang J
Tue Oct 01 00:00:00 EDT 2019
Kallikrein 12 Regulates Innate Resistance of Murine Macrophages against Mycobacterium bovis Infection by Modulating Autophagy and Apoptosis.
Cells
Sabir N,Hussain T,Liao Y,Wang J,Song Y,Shahid M,Cheng G,Mangi MH,Yao J,Yang L,Zhao D,Zhou X
Sun May 05 00:00:00 EDT 2019
A1CF-Axin2 signal axis regulates apoptosis and migration in Wilms tumor-derived cells through Wnt/β-catenin pathway.
In vitro cellular & developmental biology. Animal
Ni D,Liu J,Hu Y,Liu Y,Gu Y,Zhou Q,Xie Y
Mon Apr 01 00:00:00 EDT 2019
Exogenous regucalcin negatively regulates the progression of cervical adenocarcinoma.
Oncology letters
Li X,Huang Y,Guo S,Xie M,Bin X,Shi M,Chen A,Chen S,Wu F,Hu Q,Zhou S
Mon Jul 01 00:00:00 EDT 2019
Applying vibration in early postmenopausal osteoporosis promotes osteogenic differentiation of bone marrow-derived mesenchymal stem cells and suppresses postmenopausal osteoporosis progression.
Bioscience reports
Li H,Wu W,He X,Cao C,Yu X,Zeng Y,Li L
Mon Sep 30 00:00:00 EDT 2019
Centriolar satellites are required for efficient ciliogenesis and ciliary content regulation.
EMBO reports
Odabasi E,Gul S,Kavakli IH,Firat-Karalar EN
Sat Jun 01 00:00:00 EDT 2019
Sphingomyelin Synthase 2 Promotes Endothelial Dysfunction by Inducing Endoplasmic Reticulum Stress.
International journal of molecular sciences
Hua L,Wu N,Zhao R,He X,Liu Q,Li X,He Z,Yu L,Yan N
Wed Jun 12 00:00:00 EDT 2019
MiR-29b/Sp1/FUT4 axis modulates the malignancy of leukemia stem cells by regulating fucosylation via Wnt/β-catenin pathway in acute myeloid leukemia.
Journal of experimental & clinical cancer research : CR
Liu B,Ma H,Liu Q,Xiao Y,Pan S,Zhou H,Jia L
Thu May 16 00:00:00 EDT 2019
Scorpion Venom Analgesic Peptide, BmK AGAP Inhibits Stemness, and Epithelial-Mesenchymal Transition by Down-Regulating PTX3 in Breast Cancer.
Frontiers in oncology
Kampo S,Ahmmed B,Zhou T,Owusu L,Anabah TW,Doudou NR,Kuugbee ED,Cui Y,Lu Z,Yan Q,Wen QP
Tue Sep 19 00:00:00 EDT 2023
STRA6 exerts oncogenic role in gastric tumorigenesis by acting as a crucial target of miR-873.
Journal of experimental & clinical cancer research : CR
Lin L,Xiao J,Shi L,Chen W,Ge Y,Jiang M,Li Z,Fan H,Yang L,Xu Z
Wed Nov 06 00:00:00 EST 2019
TMEM18 inhibits osteogenic differentiation of rat bone marrow-derived mesenchymal stem cells by inactivating β-catenin.
Experimental cell research
Zhang Y,Wang H,Yin T,Liu Y,Zhou W,Fan X,Wu L,Song C,Shao J,Yang T
Tue Oct 01 00:00:00 EDT 2019
TMEM18 inhibits osteogenic differentiation of rat bone marrow-derived mesenchymal stem cells by inactivating β-catenin.
Experimental cell research
Zhang Y,Wang H,Yin T,Liu Y,Zhou W,Fan X,Wu L,Song C,Shao J,Yang T
Tue Oct 01 00:00:00 EDT 2019
miR-504 suppresses mesenchymal phenotype of glioblastoma by directly targeting the FZD7-mediated Wnt-β-catenin pathway.
Journal of experimental & clinical cancer research : CR
Liu Q,Guan Y,Li Z,Wang Y,Liu Y,Cui R,Wang Y
Fri Aug 16 00:00:00 EDT 2019
Thyroid hormone (T3) is involved in inhibiting the proliferation of newborn calf Sertoli cells via the PI3K/Akt signaling pathway in vitro.
Theriogenology
Wang C,Zheng P,Adeniran SO,Ma M,Huang F,Adegoke EO,Zhang G
Mon Jul 15 00:00:00 EDT 2019
Influence of Wilms' tumor suppressor gene WT1 on bovine Sertoli cells polarity and tight junctions via non-canonical WNT signaling pathway.
Theriogenology
Wang X,Adegoke EO,Ma M,Huang F,Zhang H,Adeniran SO,Zheng P,Zhang G
Tue Oct 15 00:00:00 EDT 2019
USP9X-mediated deubiquitination of B-cell CLL/lymphoma 9 potentiates Wnt signaling and promotes breast carcinogenesis.
The Journal of biological chemistry
Shang Z,Zhao J,Zhang Q,Cao C,Tian S,Zhang K,Liu L,Shi L,Yu N,Yang S
Fri Jun 21 00:00:00 EDT 2019
XQ-1H alleviates cerebral ischemia in mice through inhibition of apoptosis and promotion of neurogenesis in a Wnt/β-catenin signaling dependent way.
Life sciences
Xu D,Hou K,Li F,Chen S,Fang W,Li Y
Tue Oct 15 00:00:00 EDT 2019
XQ-1H alleviates cerebral ischemia in mice through inhibition of apoptosis and promotion of neurogenesis in a Wnt/β-catenin signaling dependent way.
Life sciences
Xu D,Hou K,Li F,Chen S,Fang W,Li Y
Tue Oct 15 00:00:00 EDT 2019
TRIP4 promotes tumor growth and metastasis and regulates radiosensitivity of cervical cancer by activating MAPK, PI3K/AKT, and hTERT signaling.
Cancer letters
Che Y,Li Y,Zheng F,Zou K,Li Z,Chen M,Hu S,Tian C,Yu W,Guo W,Luo M,Deng W,Zou L
Fri Jun 28 00:00:00 EDT 2019
Anti-osteoporosis effects of osteoking via reducing reactive oxygen species.
Journal of ethnopharmacology
Qin D,Zhang H,Zhang H,Sun T,Zhao H,Lee WH
Fri Nov 15 00:00:00 EST 2019
Single-cell RNA-seq reveals the invasive trajectory and molecular cascades underlying glioblastoma progression.
Molecular oncology
Pang B,Xu J,Hu J,Guo F,Wan L,Cheng M,Pang L
Sun Dec 01 00:00:00 EST 2019
BECN1 promotes the migration of NSCLC cells through regulating the ubiquitination of Vimentin.
Cell adhesion & migration
Cheng Z,Xin H,Han T
Sun Dec 01 00:00:00 EST 2019
Stimulation of endothelial progenitor cells by microRNA-31a-5p to induce endothelialization in an aneurysm neck after coil embolization by modulating the Axin1-mediated β-catenin/vascular endothelial growth factor pathway.
Journal of neurosurgery
Yu G,Liu P,Shi Y,Li S,Liu Y,Fan Z,Zhu W
Fri Aug 09 00:00:00 EDT 2019
Genome-wide screening identifies novel genes and biological processes implicated in cisplatin resistance.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Ko T,Li S
Sat Jun 01 00:00:00 EDT 2019
Betulinic Acid Suppresses Breast Cancer Metastasis by Targeting GRP78-Mediated Glycolysis and ER Stress Apoptotic Pathway.
Oxidative medicine and cellular longevity
Zheng Y,Liu P,Wang N,Wang S,Yang B,Li M,Chen J,Situ H,Xie M,Lin Y,Wang Z
Thu Feb 06 00:00:00 EST 2020
Ursolic acid promotes apoptosis and mediates transcriptional suppression of CT45A2 gene expression in non-small-cell lung carcinoma harbouring EGFR T790M mutations.
British journal of pharmacology
Yang K,Chen Y,Zhou J,Ma L,Shan Y,Cheng X,Wang Y,Zhang Z,Ji X,Chen L,Dai H,Zhu B,Li C,Tao Z,Hu X,Yin W
Sun Dec 01 00:00:00 EST 2019
Ursolic acid promotes apoptosis and mediates transcriptional suppression of CT45A2 gene expression in non-small-cell lung carcinoma harbouring EGFR T790M mutations.
British journal of pharmacology
Yang K,Chen Y,Zhou J,Ma L,Shan Y,Cheng X,Wang Y,Zhang Z,Ji X,Chen L,Dai H,Zhu B,Li C,Tao Z,Hu X,Yin W
Sun Dec 01 00:00:00 EST 2019
CLK3 Is A Direct Target Of miR-144 And Contributes To Aggressive Progression In Hepatocellular Carcinoma.
OncoTargets and therapy
Li H,Cui X,Hu Q,Chen X,Zhou P
Mon Apr 11 00:00:00 EDT 2022
Exogenous H2S mitigates myocardial fibrosis in diabetic rats through suppression of the canonical Wnt pathway.
International journal of molecular medicine
Yang R,Jia Q,Ma SF,Wang Y,Mehmood S,Chen Y
Thu Aug 01 00:00:00 EDT 2019
Knockdown of SP1/Syncytin1 axis inhibits the proliferation and metastasis through the AKT and ERK1/2 signaling pathways in non-small cell lung cancer.
Cancer medicine
Li X,Fu Y,Xia X,Zhang X,Xiao K,Zhuang X,Zhang Y
Sun Sep 01 00:00:00 EDT 2019
XIAOPI Formula Inhibits Breast Cancer Stem Cells via Suppressing Tumor-Associated Macrophages/C-X-C Motif Chemokine Ligand 1 Pathway.
Frontiers in pharmacology
Wang S,Liu X,Huang R,Zheng Y,Wang N,Yang B,Situ H,Lin Y,Wang Z
Thu Oct 01 00:00:00 EDT 2020
Overexpression of Circulating Soluble Nogo-B Improves Diabetic Kidney Disease by Protecting the Vasculature.
Diabetes
Hernandez-Diaz I,Pan J,Ricciardi CA,Bai X,Ke J,White KE,Flaquer M,Fouli GE,Argunhan F,Hayward AE,Hou FF,Mann GE,Miao RQ,Long DA,Gnudi L
Sun Sep 01 00:00:00 EDT 2019
NUDT21 inhibits bladder cancer progression through ANXA2 and LIMK2 by alternative polyadenylation.
Theranostics
Xiong M,Chen L,Zhou L,Ding Y,Kazobinka G,Chen Z,Hou T
Wed Sep 16 00:00:00 EDT 2020
O-GlcNAcylation of Thr12/Ser56 in short-form O-GlcNAc transferase (sOGT) regulates its substrate selectivity.
The Journal of biological chemistry
Liu L,Li L,Ma C,Shi Y,Liu C,Xiao Z,Zhang Y,Tian F,Gao Y,Zhang J,Ying W,Wang PG,Zhang L
Fri Nov 08 00:00:00 EST 2019
Identification of RE1-Silencing Transcription Factor as a Promoter of Metastasis in Pancreatic Cancer.
Frontiers in oncology
Jin H,Liu P,Kong L,Fei X,Gao Y,Wu T,Sun D,Tan X
Wed Sep 30 00:00:00 EDT 2020
Simvastatin promotes endothelial dysfunction by activating the Wnt/β‑catenin pathway under oxidative stress.
International journal of molecular medicine
He Z,Du X,Wu Y,Hua L,Wan L,Yan N
Tue Oct 01 00:00:00 EDT 2019
CD36 inhibits β-catenin/c-myc-mediated glycolysis through ubiquitination of GPC4 to repress colorectal tumorigenesis.
Nature communications
Fang Y,Shen ZY,Zhan YZ,Feng XC,Chen KL,Li YS,Deng HJ,Pan SM,Wu DH,Ding Y
Wed Sep 04 00:00:00 EDT 2019
Verteporfin suppresses the proliferation, epithelial-mesenchymal transition and stemness of head and neck squamous carcinoma cells via inhibiting YAP1.
Journal of Cancer
Liu K,Du S,Gao P,Zheng J
Thu Oct 01 00:00:00 EDT 2020
UPLC-Q-TOF/MS characterization of efficacy substances on osteoblasts differentiation and function in rat serum after administration of Wang-Bi tablet.
Biomedical chromatography : BMC
Wang H,Guan Y,Wu R,Lv X,Shen X,Ye G
Tue Oct 01 00:00:00 EDT 2019
Basolateral CD147 induces hepatocyte polarity loss by E-cadherin ubiquitination and degradation in hepatocellular carcinoma progress.
Hepatology (Baltimore, Md.)
Lu M,Wu J,Hao ZW,Shang YK,Xu J,Nan G,Li X,Chen ZN,Bian H
Sun Jul 01 00:00:00 EDT 2018
RAB37 interacts directly with ATG5 and promotes autophagosome formation via regulating ATG5-12-16 complex assembly.
Cell death and differentiation
Sheng Y,Song Y,Li Z,Wang Y,Lin H,Cheng H,Zhou R
Tue May 01 00:00:00 EDT 2018
RAB37 interacts directly with ATG5 and promotes autophagosome formation via regulating ATG5-12-16 complex assembly.
Cell death and differentiation
Sheng Y,Song Y,Li Z,Wang Y,Lin H,Cheng H,Zhou R
Tue May 01 00:00:00 EDT 2018
Fluoride altered rat's blood testis barrier by affecting the F-actin via IL-1α.
Chemosphere
He X,Sun Z,Manthari RK,Wu P,Wang J
Thu Nov 01 00:00:00 EDT 2018
Fluoride altered rat's blood testis barrier by affecting the F-actin via IL-1α.
Chemosphere
He X,Sun Z,Manthari RK,Wu P,Wang J
Thu Nov 01 00:00:00 EDT 2018
Circadian regulator NR1D2 regulates glioblastoma cell proliferation and motility.
Oncogene
Yu M,Li W,Wang Q,Wang Y,Lu F
Wed Aug 01 00:00:00 EDT 2018
Circadian regulator NR1D2 regulates glioblastoma cell proliferation and motility.
Oncogene
Yu M,Li W,Wang Q,Wang Y,Lu F
Wed Aug 01 00:00:00 EDT 2018
Glutathione reductase-mediated thiol oxidative stress suppresses metastasis of murine melanoma cells.
Free radical biology & medicine
Li X,Wu J,Zhang X,Chen W
Sat Dec 01 00:00:00 EST 2018
Glutathione reductase-mediated thiol oxidative stress suppresses metastasis of murine melanoma cells.
Free radical biology & medicine
Li X,Wu J,Zhang X,Chen W
Sat Dec 01 00:00:00 EST 2018
The gastric mucosal protective effects of astragaloside IV in mnng-induced GPL rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Cai T,Zhang C,Zhao Z,Li S,Cai H,Chen X,Cai D,Liu W,Yan Y,Xie K,Pan H,Zeng X
Wed Aug 01 00:00:00 EDT 2018
The gastric mucosal protective effects of astragaloside IV in mnng-induced GPL rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Cai T,Zhang C,Zhao Z,Li S,Cai H,Chen X,Cai D,Liu W,Yan Y,Xie K,Pan H,Zeng X
Wed Aug 01 00:00:00 EDT 2018
Sodium Selenite inhibits mitophagy, downregulation and mislocalization of blood-testis barrier proteins of bovine Sertoli cell exposed to microcystin-leucine arginine (MC-LR) via TLR4/NF-kB and mitochondrial signaling pathways blockage.
Ecotoxicology and environmental safety
Adegoke EO,Xue W,Machebe NS,Adeniran SO,Hao W,Chen W,Han Z,Guixue Z,Peng Z
Sun Dec 30 00:00:00 EST 2018
A novel Anti-Cancer Stem Cells compound optimized from the natural symplostatin 4 scaffold inhibits Wnt/β-catenin signaling pathway.
European journal of medicinal chemistry
Liu S,Gao X,Zhang L,Qin S,Wei M,Liu N,Zhao R,Li B,Meng Y,Lin G,Lu C,Liu X,Xie M,Liu T,Zhou H,Qi M,Yang G,Yang C
Sun Aug 05 00:00:00 EDT 2018
A novel Anti-Cancer Stem Cells compound optimized from the natural symplostatin 4 scaffold inhibits Wnt/β-catenin signaling pathway.
European journal of medicinal chemistry
Liu S,Gao X,Zhang L,Qin S,Wei M,Liu N,Zhao R,Li B,Meng Y,Lin G,Lu C,Liu X,Xie M,Liu T,Zhou H,Qi M,Yang G,Yang C
Sun Aug 05 00:00:00 EDT 2018
Krüppel-like factor 4 promotes c-Met amplification-mediated gefitinib resistance in non-small-cell lung cancer.
Cancer science
Feng W,Xie Q,Liu S,Ji Y,Li C,Wang C,Jin L
Fri Jun 01 00:00:00 EDT 2018
Krüppel-like factor 4 promotes c-Met amplification-mediated gefitinib resistance in non-small-cell lung cancer.
Cancer science
Feng W,Xie Q,Liu S,Ji Y,Li C,Wang C,Jin L
Fri Jun 01 00:00:00 EDT 2018
Phenethyl isothiocyanate inhibits colorectal cancer stem cells by suppressing Wnt/β-catenin pathway.
Phytotherapy research : PTR
Chen Y,Li Y,Wang XQ,Meng Y,Zhang Q,Zhu JY,Chen JQ,Cao WS,Wang XQ,Xie CF,Li XT,Geng SS,Wu JS,Zhong CY,Han HY
Sat Dec 01 00:00:00 EST 2018
Phenethyl isothiocyanate inhibits colorectal cancer stem cells by suppressing Wnt/β-catenin pathway.
Phytotherapy research : PTR
Chen Y,Li Y,Wang XQ,Meng Y,Zhang Q,Zhu JY,Chen JQ,Cao WS,Wang XQ,Xie CF,Li XT,Geng SS,Wu JS,Zhong CY,Han HY
Sat Dec 01 00:00:00 EST 2018
Down-regulation of long non-coding RNA ITGB2-AS1 inhibits osteosarcoma proliferation and metastasis by repressing Wnt/β-catenin signalling and predicts favourable prognosis.
Artificial cells, nanomedicine, and biotechnology
Dai J,Xu LJ,Han GD,Jiang HT,Sun HL,Zhu GT,Tang XM
Tue Jun 18 00:00:00 EDT 2019
Down-regulation of long non-coding RNA ITGB2-AS1 inhibits osteosarcoma proliferation and metastasis by repressing Wnt/β-catenin signalling and predicts favourable prognosis.
Artificial cells, nanomedicine, and biotechnology
Dai J,Xu LJ,Han GD,Jiang HT,Sun HL,Zhu GT,Tang XM
Tue Jun 18 00:00:00 EDT 2019
AMPK/GSK3β/β-catenin cascade-triggered overexpression of CEMIP promotes migration and invasion in anoikis-resistant prostate cancer cells by enhancing metabolic reprogramming.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhang P,Song Y,Sun Y,Li X,Chen L,Yang L,Xing Y
Sun Jul 01 00:00:00 EDT 2018
AMPK/GSK3β/β-catenin cascade-triggered overexpression of CEMIP promotes migration and invasion in anoikis-resistant prostate cancer cells by enhancing metabolic reprogramming.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhang P,Song Y,Sun Y,Li X,Chen L,Yang L,Xing Y
Sun Jul 01 00:00:00 EDT 2018
Microcystin-leucine arginine (MC-LR) induced inflammatory response in bovine sertoli cell via TLR4/NF-kB signaling pathway.
Environmental toxicology and pharmacology
Adegoke EO,Wang C,Machebe NS,Wang X,Wang H,Adeniran SO,Zhang H,Zheng P,Zhang G
Mon Oct 01 00:00:00 EDT 2018
Microcystin-leucine arginine (MC-LR) induced inflammatory response in bovine sertoli cell via TLR4/NF-kB signaling pathway.
Environmental toxicology and pharmacology
Adegoke EO,Wang C,Machebe NS,Wang X,Wang H,Adeniran SO,Zhang H,Zheng P,Zhang G
Mon Oct 01 00:00:00 EDT 2018
Anticancer activity and potential mechanisms of 1C, a ginseng saponin derivative, on prostate cancer cells.
Journal of ginseng research
Wang X,Su GY,Zhao C,Qu FZ,Wang P,Zhao YQ
Sun Apr 01 00:00:00 EDT 2018
Anticancer activity and potential mechanisms of 1C, a ginseng saponin derivative, on prostate cancer cells.
Journal of ginseng research
Wang X,Su GY,Zhao C,Qu FZ,Wang P,Zhao YQ
Sun Apr 01 00:00:00 EDT 2018
Octreotide attenuates hepatic fibrosis and hepatic stellate cells proliferation and activation by inhibiting Wnt/β-catenin signaling pathway, c-Myc and cyclin D1.
International immunopharmacology
Zhang C,Liu XQ,Sun HN,Meng XM,Bao YW,Zhang HP,Pan FM,Zhang C
Mon Oct 01 00:00:00 EDT 2018
Octreotide attenuates hepatic fibrosis and hepatic stellate cells proliferation and activation by inhibiting Wnt/β-catenin signaling pathway, c-Myc and cyclin D1.
International immunopharmacology
Zhang C,Liu XQ,Sun HN,Meng XM,Bao YW,Zhang HP,Pan FM,Zhang C
Mon Oct 01 00:00:00 EDT 2018
Octreotide attenuates hepatic fibrosis and hepatic stellate cells proliferation and activation by inhibiting Wnt/β-catenin signaling pathway, c-Myc and cyclin D1.
International immunopharmacology
Zhang C,Liu XQ,Sun HN,Meng XM,Bao YW,Zhang HP,Pan FM,Zhang C
Mon Oct 01 00:00:00 EDT 2018
Octreotide attenuates hepatic fibrosis and hepatic stellate cells proliferation and activation by inhibiting Wnt/β-catenin signaling pathway, c-Myc and cyclin D1.
International immunopharmacology
Zhang C,Liu XQ,Sun HN,Meng XM,Bao YW,Zhang HP,Pan FM,Zhang C
Mon Oct 01 00:00:00 EDT 2018
Ionizing Radiation Blocks Hair Cell Regeneration in Zebrafish Lateral Line Neuromasts by Preventing Wnt Signaling.
Molecular neurobiology
Li R,Liao G,Yin G,Wang B,Yan M,Lin X,Zhang W,Chen X,Du S,Yuan Y
Thu Feb 01 00:00:00 EST 2018
Ionizing Radiation Blocks Hair Cell Regeneration in Zebrafish Lateral Line Neuromasts by Preventing Wnt Signaling.
Molecular neurobiology
Li R,Liao G,Yin G,Wang B,Yan M,Lin X,Zhang W,Chen X,Du S,Yuan Y
Thu Feb 01 00:00:00 EST 2018
Retigabine ameliorates acute stress-induced impairment of spatial memory retrieval through regulating USP2 signaling pathways in hippocampal CA1 area.
Neuropharmacology
Li C,Zhang J,Xu H,Chang M,Lv C,Xue W,Song Z,Zhang L,Zhang X,Tian X
Fri Jun 01 00:00:00 EDT 2018
Retigabine ameliorates acute stress-induced impairment of spatial memory retrieval through regulating USP2 signaling pathways in hippocampal CA1 area.
Neuropharmacology
Li C,Zhang J,Xu H,Chang M,Lv C,Xue W,Song Z,Zhang L,Zhang X,Tian X
Fri Jun 01 00:00:00 EDT 2018
Wenyang Huazhuo Tongluo formula inhibits fibrosis via suppressing Wnt/β-catenin signaling pathway in a Bleomycin-induced systemic sclerosis mouse model.
Chinese medicine
Wang Q,Zang W,Han L,Yang L,Ye S,Ouyang J,Zhang C,Bi Y,Zhang C,Bian H
Fri Apr 08 00:00:00 EDT 2022
Wenyang Huazhuo Tongluo formula inhibits fibrosis via suppressing Wnt/β-catenin signaling pathway in a Bleomycin-induced systemic sclerosis mouse model.
Chinese medicine
Wang Q,Zang W,Han L,Yang L,Ye S,Ouyang J,Zhang C,Bi Y,Zhang C,Bian H
Fri Apr 08 00:00:00 EDT 2022
Diallyl Trisulfide inhibits breast cancer stem cells via suppression of Wnt/β-catenin pathway.
Journal of cellular biochemistry
Li X,Meng Y,Xie C,Zhu J,Wang X,Li Y,Geng S,Wu J,Zhong C,Li M
Tue May 01 00:00:00 EDT 2018
Diallyl Trisulfide inhibits breast cancer stem cells via suppression of Wnt/β-catenin pathway.
Journal of cellular biochemistry
Li X,Meng Y,Xie C,Zhu J,Wang X,Li Y,Geng S,Wu J,Zhong C,Li M
Tue May 01 00:00:00 EDT 2018
S100A4-MYH9 Axis Promote Migration and Invasion of Gastric Cancer Cells by Inducing TGF-β-Mediated Epithelial-Mesenchymal Transition.
Journal of Cancer
Li F,Shi J,Xu Z,Yao X,Mou T,Yu J,Liu H,Li G
Wed Sep 30 00:00:00 EDT 2020
S100A4-MYH9 Axis Promote Migration and Invasion of Gastric Cancer Cells by Inducing TGF-β-Mediated Epithelial-Mesenchymal Transition.
Journal of Cancer
Li F,Shi J,Xu Z,Yao X,Mou T,Yu J,Liu H,Li G
Wed Sep 30 00:00:00 EDT 2020
S100A4-MYH9 Axis Promote Migration and Invasion of Gastric Cancer Cells by Inducing TGF-β-Mediated Epithelial-Mesenchymal Transition.
Journal of Cancer
Li F,Shi J,Xu Z,Yao X,Mou T,Yu J,Liu H,Li G
Wed Sep 30 00:00:00 EDT 2020
S100A4-MYH9 Axis Promote Migration and Invasion of Gastric Cancer Cells by Inducing TGF-β-Mediated Epithelial-Mesenchymal Transition.
Journal of Cancer
Li F,Shi J,Xu Z,Yao X,Mou T,Yu J,Liu H,Li G
Wed Sep 30 00:00:00 EDT 2020
P21-activated kinase 7 (PAK7) interacts with and activates Wnt/β-catenin signaling pathway in breast cancer.
Journal of Cancer
Li K,Xu X,He Y,Tian Y,Pan W,Xu L,Ma Y,Gao Y,Gao J,Qi Y,Wei L,Zhang J
Sun Nov 12 00:00:00 EST 2023
P21-activated kinase 7 (PAK7) interacts with and activates Wnt/β-catenin signaling pathway in breast cancer.
Journal of Cancer
Li K,Xu X,He Y,Tian Y,Pan W,Xu L,Ma Y,Gao Y,Gao J,Qi Y,Wei L,Zhang J
Sun Nov 12 00:00:00 EST 2023
RHBDF1 regulates APC-mediated stimulation of the epithelial-to-mesenchymal transition and proliferation of colorectal cancer cells in part via the Wnt/β-catenin signalling pathway.
Experimental cell research
Yuan H,Wei R,Xiao Y,Song Y,Wang J,Yu H,Fang T,Xu W,Mao S
Sun Jul 01 00:00:00 EDT 2018
RHBDF1 regulates APC-mediated stimulation of the epithelial-to-mesenchymal transition and proliferation of colorectal cancer cells in part via the Wnt/β-catenin signalling pathway.
Experimental cell research
Yuan H,Wei R,Xiao Y,Song Y,Wang J,Yu H,Fang T,Xu W,Mao S
Sun Jul 01 00:00:00 EDT 2018
A Novel Cx50 Insert Mutation from a Chinese Congenital Cataract Family Impairs Its Cellular Membrane Localization and Function.
DNA and cell biology
Cui X,Zhou Z,Zhu K,Feng R,Han J,Li M,Wang S,Li J,Zhang J,Jiang Q,Zhang W,Mu H,Liu Y,Hu Y
Tue May 01 00:00:00 EDT 2018
A Novel Cx50 Insert Mutation from a Chinese Congenital Cataract Family Impairs Its Cellular Membrane Localization and Function.
DNA and cell biology
Cui X,Zhou Z,Zhu K,Feng R,Han J,Li M,Wang S,Li J,Zhang J,Jiang Q,Zhang W,Mu H,Liu Y,Hu Y
Tue May 01 00:00:00 EDT 2018
miR-21 modulates prostaglandin signaling and promotes gastric tumorigenesis by targeting 15-PGDH.
Biochemical and biophysical research communications
Li L,Wang X,Li W,Yang L,Liu R,Zeng R,Wu Y,Shou T
Mon Jan 01 00:00:00 EST 2018
miR-21 modulates prostaglandin signaling and promotes gastric tumorigenesis by targeting 15-PGDH.
Biochemical and biophysical research communications
Li L,Wang X,Li W,Yang L,Liu R,Zeng R,Wu Y,Shou T
Mon Jan 01 00:00:00 EST 2018
Exosomes from C2C12 myoblasts enhance osteogenic differentiation of MC3T3-E1 pre-osteoblasts by delivering miR-27a-3p.
Biochemical and biophysical research communications
Xu Q,Cui Y,Luan J,Zhou X,Li H,Han J
Sun Mar 25 00:00:00 EDT 2018
Exosomes from C2C12 myoblasts enhance osteogenic differentiation of MC3T3-E1 pre-osteoblasts by delivering miR-27a-3p.
Biochemical and biophysical research communications
Xu Q,Cui Y,Luan J,Zhou X,Li H,Han J
Sun Mar 25 00:00:00 EDT 2018
Exosomes from C2C12 myoblasts enhance osteogenic differentiation of MC3T3-E1 pre-osteoblasts by delivering miR-27a-3p.
Biochemical and biophysical research communications
Xu Q,Cui Y,Luan J,Zhou X,Li H,Han J
Sun Mar 25 00:00:00 EDT 2018
Exosomes from C2C12 myoblasts enhance osteogenic differentiation of MC3T3-E1 pre-osteoblasts by delivering miR-27a-3p.
Biochemical and biophysical research communications
Xu Q,Cui Y,Luan J,Zhou X,Li H,Han J
Sun Mar 25 00:00:00 EDT 2018
Wnt/β-catenin signaling mediates the suppressive effects of diallyl trisulfide on colorectal cancer stem cells.
Cancer chemotherapy and pharmacology
Zhang Q,Li XT,Chen Y,Chen JQ,Zhu JY,Meng Y,Wang XQ,Li Y,Geng SS,Xie CF,Wu JS,Zhong CY,Han HY
Fri Jun 01 00:00:00 EDT 2018
Wnt/β-catenin signaling mediates the suppressive effects of diallyl trisulfide on colorectal cancer stem cells.
Cancer chemotherapy and pharmacology
Zhang Q,Li XT,Chen Y,Chen JQ,Zhu JY,Meng Y,Wang XQ,Li Y,Geng SS,Xie CF,Wu JS,Zhong CY,Han HY
Fri Jun 01 00:00:00 EDT 2018
β-catenin, leucine-rich repeat-containing G protein-coupled receptor 5 and GATA-binding factor 6 are associated with the normal mucosa-adenoma-adenocarcinoma sequence of colorectal tumorigenesis.
Oncology letters
Yang B,Mao L,Li Y,Li Q,Li X,Zhang Y,Zhai Z
Thu Feb 01 00:00:00 EST 2018
β-catenin, leucine-rich repeat-containing G protein-coupled receptor 5 and GATA-binding factor 6 are associated with the normal mucosa-adenoma-adenocarcinoma sequence of colorectal tumorigenesis.
Oncology letters
Yang B,Mao L,Li Y,Li Q,Li X,Zhang Y,Zhai Z
Thu Feb 01 00:00:00 EST 2018
Protective effects of 2,3,5,4-tetrahydroxystilbene-2-o-β-D-glucoside against osteoporosis: Current knowledge and proposed mechanisms.
International journal of rheumatic diseases
Zhang J,Li S,Wei L,Peng Y,Zheng Z,Xue J,Cao Y,Wang B,Du J
Wed Aug 01 00:00:00 EDT 2018
Protective effects of 2,3,5,4-tetrahydroxystilbene-2-o-β-D-glucoside against osteoporosis: Current knowledge and proposed mechanisms.
International journal of rheumatic diseases
Zhang J,Li S,Wei L,Peng Y,Zheng Z,Xue J,Cao Y,Wang B,Du J
Wed Aug 01 00:00:00 EDT 2018
Silencing of lncRNA ZFAS1 inhibits malignancies by blocking Wnt/β-catenin signaling in gastric cancer cells.
Bioscience, biotechnology, and biochemistry
Xu W,He L,Li Y,Tan Y,Zhang F,Xu H
Thu Mar 01 00:00:00 EST 2018
Silencing of lncRNA ZFAS1 inhibits malignancies by blocking Wnt/β-catenin signaling in gastric cancer cells.
Bioscience, biotechnology, and biochemistry
Xu W,He L,Li Y,Tan Y,Zhang F,Xu H
Thu Mar 01 00:00:00 EST 2018
Silencing of lncRNA ZFAS1 inhibits malignancies by blocking Wnt/β-catenin signaling in gastric cancer cells.
Bioscience, biotechnology, and biochemistry
Xu W,He L,Li Y,Tan Y,Zhang F,Xu H
Thu Mar 01 00:00:00 EST 2018
Silencing of lncRNA ZFAS1 inhibits malignancies by blocking Wnt/β-catenin signaling in gastric cancer cells.
Bioscience, biotechnology, and biochemistry
Xu W,He L,Li Y,Tan Y,Zhang F,Xu H
Thu Mar 01 00:00:00 EST 2018
Silencing of CEMIP suppresses Wnt/β-catenin/Snail signaling transduction and inhibits EMT program of colorectal cancer cells.
Acta histochemica
Liang G,Fang X,Yang Y,Song Y
Mon Jan 01 00:00:00 EST 2018
Silencing of CEMIP suppresses Wnt/β-catenin/Snail signaling transduction and inhibits EMT program of colorectal cancer cells.
Acta histochemica
Liang G,Fang X,Yang Y,Song Y
Mon Jan 01 00:00:00 EST 2018
Silencing of CEMIP suppresses Wnt/β-catenin/Snail signaling transduction and inhibits EMT program of colorectal cancer cells.
Acta histochemica
Liang G,Fang X,Yang Y,Song Y
Mon Jan 01 00:00:00 EST 2018
Silencing of CEMIP suppresses Wnt/β-catenin/Snail signaling transduction and inhibits EMT program of colorectal cancer cells.
Acta histochemica
Liang G,Fang X,Yang Y,Song Y
Mon Jan 01 00:00:00 EST 2018
Wnt signaling pathway correlates with ossification of the spinal ligament: A microRNA array and immunohistochemical study.
Journal of orthopaedic science : official journal of the Japanese Orthopaedic Association
Yayama T,Mori K,Okumura N,Nishizawa K,Kumagai K,Nakamura A,Imai S
Mon Jan 01 00:00:00 EST 2018
Wnt signaling pathway correlates with ossification of the spinal ligament: A microRNA array and immunohistochemical study.
Journal of orthopaedic science : official journal of the Japanese Orthopaedic Association
Yayama T,Mori K,Okumura N,Nishizawa K,Kumagai K,Nakamura A,Imai S
Mon Jan 01 00:00:00 EST 2018
Reduced graphene oxide triggered epithelial-mesenchymal transition in A549 cells.
Scientific reports
Liao Y,Wang W,Huang X,Sun Y,Tian S,Cai P
Fri Oct 12 00:00:00 EDT 2018
Reduced graphene oxide triggered epithelial-mesenchymal transition in A549 cells.
Scientific reports
Liao Y,Wang W,Huang X,Sun Y,Tian S,Cai P
Fri Oct 12 00:00:00 EDT 2018
High Concentration of Aspirin Induces Apoptosis in Rat Tendon Stem Cells via Inhibition of the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Y,Tang H,He G,Shi Y,Kang X,Lyu J,Zhou M,Zhu M,Zhang J,Tang K
Tue Dec 11 00:00:00 EST 2018
High Concentration of Aspirin Induces Apoptosis in Rat Tendon Stem Cells via Inhibition of the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Y,Tang H,He G,Shi Y,Kang X,Lyu J,Zhou M,Zhu M,Zhang J,Tang K
Tue Dec 11 00:00:00 EST 2018
High Concentration of Aspirin Induces Apoptosis in Rat Tendon Stem Cells via Inhibition of the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Y,Tang H,He G,Shi Y,Kang X,Lyu J,Zhou M,Zhu M,Zhang J,Tang K
Tue Dec 11 00:00:00 EST 2018
High Concentration of Aspirin Induces Apoptosis in Rat Tendon Stem Cells via Inhibition of the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wang Y,Tang H,He G,Shi Y,Kang X,Lyu J,Zhou M,Zhu M,Zhang J,Tang K
Tue Dec 11 00:00:00 EST 2018
Periostin secreted by cancer-associated fibroblasts promotes cancer stemness in head and neck cancer by activating protein tyrosine kinase 7.
Cell death & disease
Yu B,Wu K,Wang X,Zhang J,Wang L,Jiang Y,Zhu X,Chen W,Yan M
Mon Oct 22 00:00:00 EDT 2018
Periostin secreted by cancer-associated fibroblasts promotes cancer stemness in head and neck cancer by activating protein tyrosine kinase 7.
Cell death & disease
Yu B,Wu K,Wang X,Zhang J,Wang L,Jiang Y,Zhu X,Chen W,Yan M
Mon Oct 22 00:00:00 EDT 2018
Periostin secreted by cancer-associated fibroblasts promotes cancer stemness in head and neck cancer by activating protein tyrosine kinase 7.
Cell death & disease
Yu B,Wu K,Wang X,Zhang J,Wang L,Jiang Y,Zhu X,Chen W,Yan M
Mon Oct 22 00:00:00 EDT 2018
Periostin secreted by cancer-associated fibroblasts promotes cancer stemness in head and neck cancer by activating protein tyrosine kinase 7.
Cell death & disease
Yu B,Wu K,Wang X,Zhang J,Wang L,Jiang Y,Zhu X,Chen W,Yan M
Mon Oct 22 00:00:00 EDT 2018
Periostin secreted by cancer-associated fibroblasts promotes cancer stemness in head and neck cancer by activating protein tyrosine kinase 7.
Cell death & disease
Yu B,Wu K,Wang X,Zhang J,Wang L,Jiang Y,Zhu X,Chen W,Yan M
Mon Oct 22 00:00:00 EDT 2018
MicroRNA-664-5p promotes myoblast proliferation and inhibits myoblast differentiation by targeting serum response factor and Wnt1.
The Journal of biological chemistry
Cai R,Qimuge N,Ma M,Wang Y,Tang G,Zhang Q,Sun Y,Chen X,Yu T,Dong W,Yang G,Pang W
Fri Dec 14 00:00:00 EST 2018
MicroRNA-664-5p promotes myoblast proliferation and inhibits myoblast differentiation by targeting serum response factor and Wnt1.
The Journal of biological chemistry
Cai R,Qimuge N,Ma M,Wang Y,Tang G,Zhang Q,Sun Y,Chen X,Yu T,Dong W,Yang G,Pang W
Fri Dec 14 00:00:00 EST 2018
Psoralen induced cell cycle arrest by modulating Wnt/β-catenin pathway in breast cancer cells.
Scientific reports
Wang X,Xu C,Hua Y,Cheng K,Zhang Y,Liu J,Han Y,Liu S,Zhang G,Xu S,Yang Z
Tue Sep 18 00:00:00 EDT 2018
Psoralen induced cell cycle arrest by modulating Wnt/β-catenin pathway in breast cancer cells.
Scientific reports
Wang X,Xu C,Hua Y,Cheng K,Zhang Y,Liu J,Han Y,Liu S,Zhang G,Xu S,Yang Z
Tue Sep 18 00:00:00 EDT 2018
Psoralen induced cell cycle arrest by modulating Wnt/β-catenin pathway in breast cancer cells.
Scientific reports
Wang X,Xu C,Hua Y,Cheng K,Zhang Y,Liu J,Han Y,Liu S,Zhang G,Xu S,Yang Z
Tue Sep 18 00:00:00 EDT 2018
Psoralen induced cell cycle arrest by modulating Wnt/β-catenin pathway in breast cancer cells.
Scientific reports
Wang X,Xu C,Hua Y,Cheng K,Zhang Y,Liu J,Han Y,Liu S,Zhang G,Xu S,Yang Z
Tue Sep 18 00:00:00 EDT 2018
Autophagy promotes metastasis and glycolysis by upregulating MCT1 expression and Wnt/β-catenin signaling pathway activation in hepatocellular carcinoma cells.
Journal of experimental & clinical cancer research : CR
Fan Q,Yang L,Zhang X,Ma Y,Li Y,Dong L,Zong Z,Hua X,Su D,Li H,Liu J
Fri Jan 19 00:00:00 EST 2018
Autophagy promotes metastasis and glycolysis by upregulating MCT1 expression and Wnt/β-catenin signaling pathway activation in hepatocellular carcinoma cells.
Journal of experimental & clinical cancer research : CR
Fan Q,Yang L,Zhang X,Ma Y,Li Y,Dong L,Zong Z,Hua X,Su D,Li H,Liu J
Fri Jan 19 00:00:00 EST 2018
A Primate-Specific Isoform of PLEKHG6 Regulates Neurogenesis and Neuronal Migration.
Cell reports
O'Neill AC,Kyrousi C,Klaus J,Leventer RJ,Kirk EP,Fry A,Pilz DT,Morgan T,Jenkins ZA,Drukker M,Berkovic SF,Scheffer IE,Guerrini R,Markie DM,Götz M,Cappello S,Robertson SP
Tue Dec 04 00:00:00 EST 2018
A Primate-Specific Isoform of PLEKHG6 Regulates Neurogenesis and Neuronal Migration.
Cell reports
O'Neill AC,Kyrousi C,Klaus J,Leventer RJ,Kirk EP,Fry A,Pilz DT,Morgan T,Jenkins ZA,Drukker M,Berkovic SF,Scheffer IE,Guerrini R,Markie DM,Götz M,Cappello S,Robertson SP
Tue Dec 04 00:00:00 EST 2018
Baicalin increases hair follicle development by increasing canonical Wnt/β‑catenin signaling and activating dermal papillar cells in mice.
International journal of molecular medicine
Xing F,Yi WJ,Miao F,Su MY,Lei TC
Sun Apr 01 00:00:00 EDT 2018
Expression and prognostic significance of TRPV6 in the development and progression of pancreatic cancer.
Oncology reports
Song H,Dong M,Zhou J,Sheng W,Li X,Gao W
Thu Mar 01 00:00:00 EST 2018
Phosphoglycerate Mutase 1 (PGAM1) Promotes Pancreatic Ductal Adenocarcinoma (PDAC) Metastasis by Acting as a Novel Downstream Target of the PI3K/Akt/mTOR Pathway.
Oncology research
Liu X,Tan X,Liu P,Wu Y,Qian S,Zhang X
Thu Aug 23 00:00:00 EDT 2018
Upregulated IQUB promotes cell proliferation and migration via activating Akt/GSK3β/β-catenin signaling pathway in breast cancer.
Cancer medicine
Li K,Ma YB,Zhang Z,Tian YH,Xu XL,He YQ,Xu L,Gao Y,Pan WT,Song WJ,He X,Wei L
Wed Aug 01 00:00:00 EDT 2018
Upregulated IQUB promotes cell proliferation and migration via activating Akt/GSK3β/β-catenin signaling pathway in breast cancer.
Cancer medicine
Li K,Ma YB,Zhang Z,Tian YH,Xu XL,He YQ,Xu L,Gao Y,Pan WT,Song WJ,He X,Wei L
Wed Aug 01 00:00:00 EDT 2018
Resveratrol inhibits the growth of gastric cancer via the Wnt/β-catenin pathway.
Oncology letters
Dai H,Deng HB,Wang YH,Guo JJ
Wed Aug 01 00:00:00 EDT 2018
Synthesis and Discovery Novel Anti-Cancer Stem Cells Compounds Derived from the Natural Triterpenoic Acids.
Journal of medicinal chemistry
Liu X,Li B,Zhang Z,Wei Y,Xu Z,Qin S,Liu N,Zhao R,Peng J,Yang G,Qi M,Liu T,Xie M,Liu S,Gao X,Lu C,Zhu L,Long X,Kang H,Sun T,Zhou H,Wei M,Yang G,Yang C
Thu Dec 13 00:00:00 EST 2018
Synergistic effect of strontium and silicon in strontium-substituted sub-micron bioactive glass for enhanced osteogenesis.
Materials science & engineering. C, Materials for biological applications
Zhang W,Huang D,Zhao F,Gao W,Sun L,Li X,Chen X
Wed Aug 01 00:00:00 EDT 2018
The PIK3CA E542K and E545K mutations promote glycolysis and proliferation via induction of the β-catenin/SIRT3 signaling pathway in cervical cancer.
Journal of hematology & oncology
Jiang W,He T,Liu S,Zheng Y,Xiang L,Pei X,Wang Z,Yang H
Fri Dec 14 00:00:00 EST 2018
The PIK3CA E542K and E545K mutations promote glycolysis and proliferation via induction of the β-catenin/SIRT3 signaling pathway in cervical cancer.
Journal of hematology & oncology
Jiang W,He T,Liu S,Zheng Y,Xiang L,Pei X,Wang Z,Yang H
Fri Dec 14 00:00:00 EST 2018
The PIK3CA E542K and E545K mutations promote glycolysis and proliferation via induction of the β-catenin/SIRT3 signaling pathway in cervical cancer.
Journal of hematology & oncology
Jiang W,He T,Liu S,Zheng Y,Xiang L,Pei X,Wang Z,Yang H
Fri Dec 14 00:00:00 EST 2018
A CREB1/miR-433 reciprocal feedback loop modulates proliferation and metastasis in colorectal cancer.
Aging
Yan L,You WQ,Sheng NQ,Gong JF,Hu LD,Tan GW,Chen HQ,Wang ZG
Thu Dec 06 00:00:00 EST 2018
A CREB1/miR-433 reciprocal feedback loop modulates proliferation and metastasis in colorectal cancer.
Aging
Yan L,You WQ,Sheng NQ,Gong JF,Hu LD,Tan GW,Chen HQ,Wang ZG
Thu Dec 06 00:00:00 EST 2018
HIF-2α promotes conversion to a stem cell phenotype and induces chemoresistance in breast cancer cells by activating Wnt and Notch pathways.
Journal of experimental & clinical cancer research : CR
Yan Y,Liu F,Han L,Zhao L,Chen J,Olopade OI,He M,Wei M
Fri Oct 19 00:00:00 EDT 2018
HIF-2α promotes conversion to a stem cell phenotype and induces chemoresistance in breast cancer cells by activating Wnt and Notch pathways.
Journal of experimental & clinical cancer research : CR
Yan Y,Liu F,Han L,Zhao L,Chen J,Olopade OI,He M,Wei M
Fri Oct 19 00:00:00 EDT 2018
Carcinoma associated fibroblasts (CAFs) promote breast cancer motility by suppressing mammalian Diaphanous-related formin-2 (mDia2).
PloS one
Dvorak KM,Pettee KM,Rubinic-Minotti K,Su R,Nestor-Kalinoski A,Eisenmann KM
Tue Jul 24 00:00:00 EDT 2018
Carcinoma associated fibroblasts (CAFs) promote breast cancer motility by suppressing mammalian Diaphanous-related formin-2 (mDia2).
PloS one
Dvorak KM,Pettee KM,Rubinic-Minotti K,Su R,Nestor-Kalinoski A,Eisenmann KM
Tue Jul 24 00:00:00 EDT 2018
Mechanism of prolactin inhibition of miR-135b via methylation in goat mammary epithelial cells.
Journal of cellular physiology
Chen Z,Luo J,Zhang C,Ma Y,Sun S,Zhang T,Loor JJ
Mon Jan 01 00:00:00 EST 2018
Integrin-β5, a miR-185-targeted gene, promotes hepatocellular carcinoma tumorigenesis by regulating β-catenin stability.
Journal of experimental & clinical cancer research : CR
Lin Z,He R,Luo H,Lu C,Ning Z,Wu Y,Han C,Tan G,Wang Z
Wed Jan 31 00:00:00 EST 2018
Integrin-β5, a miR-185-targeted gene, promotes hepatocellular carcinoma tumorigenesis by regulating β-catenin stability.
Journal of experimental & clinical cancer research : CR
Lin Z,He R,Luo H,Lu C,Ning Z,Wu Y,Han C,Tan G,Wang Z
Wed Jan 31 00:00:00 EST 2018
Overexpression of RCC2 Enhances Cell Motility and Promotes Tumor Metastasis in Lung Adenocarcinoma by Inducing Epithelial-Mesenchymal Transition.
Clinical cancer research : an official journal of the American Association for Cancer Research
Pang B,Wu N,Guan R,Pang L,Li X,Li S,Tang L,Guo Y,Chen J,Sun D,Sun H,Dai J,Bai J,Ji G,Liu P,Liu A,Wang Q,Xiao S,Fu S,Jin Y
Fri Sep 15 00:00:00 EDT 2017
Overexpression of RCC2 Enhances Cell Motility and Promotes Tumor Metastasis in Lung Adenocarcinoma by Inducing Epithelial-Mesenchymal Transition.
Clinical cancer research : an official journal of the American Association for Cancer Research
Pang B,Wu N,Guan R,Pang L,Li X,Li S,Tang L,Guo Y,Chen J,Sun D,Sun H,Dai J,Bai J,Ji G,Liu P,Liu A,Wang Q,Xiao S,Fu S,Jin Y
Fri Sep 15 00:00:00 EDT 2017
Overexpression of RCC2 Enhances Cell Motility and Promotes Tumor Metastasis in Lung Adenocarcinoma by Inducing Epithelial-Mesenchymal Transition.
Clinical cancer research : an official journal of the American Association for Cancer Research
Pang B,Wu N,Guan R,Pang L,Li X,Li S,Tang L,Guo Y,Chen J,Sun D,Sun H,Dai J,Bai J,Ji G,Liu P,Liu A,Wang Q,Xiao S,Fu S,Jin Y
Fri Sep 15 00:00:00 EDT 2017
Overexpression of RCC2 Enhances Cell Motility and Promotes Tumor Metastasis in Lung Adenocarcinoma by Inducing Epithelial-Mesenchymal Transition.
Clinical cancer research : an official journal of the American Association for Cancer Research
Pang B,Wu N,Guan R,Pang L,Li X,Li S,Tang L,Guo Y,Chen J,Sun D,Sun H,Dai J,Bai J,Ji G,Liu P,Liu A,Wang Q,Xiao S,Fu S,Jin Y
Fri Sep 15 00:00:00 EDT 2017
Dihydroartemisin inhibits glioma invasiveness via a ROS to P53 to β-catenin signaling.
Pharmacological research
Que Z,Wang P,Hu Y,Xue Y,Liu X,Qu C,Ma J,Liu Y
Mon May 01 00:00:00 EDT 2017
Dihydroartemisin inhibits glioma invasiveness via a ROS to P53 to β-catenin signaling.
Pharmacological research
Que Z,Wang P,Hu Y,Xue Y,Liu X,Qu C,Ma J,Liu Y
Mon May 01 00:00:00 EDT 2017
Dihydroartemisin inhibits glioma invasiveness via a ROS to P53 to β-catenin signaling.
Pharmacological research
Que Z,Wang P,Hu Y,Xue Y,Liu X,Qu C,Ma J,Liu Y
Mon May 01 00:00:00 EDT 2017
Dihydroartemisin inhibits glioma invasiveness via a ROS to P53 to β-catenin signaling.
Pharmacological research
Que Z,Wang P,Hu Y,Xue Y,Liu X,Qu C,Ma J,Liu Y
Mon May 01 00:00:00 EDT 2017
Transducin (β)-like 1 X-linked receptor 1 promotes gastric cancer progression via the ERK1/2 pathway.
Oncogene
Zhou Q,Wang X,Yu Z,Wu X,Chen X,Li J,Zhu Z,Liu B,Su L
Thu Mar 30 00:00:00 EDT 2017
Transducin (β)-like 1 X-linked receptor 1 promotes gastric cancer progression via the ERK1/2 pathway.
Oncogene
Zhou Q,Wang X,Yu Z,Wu X,Chen X,Li J,Zhu Z,Liu B,Su L
Thu Mar 30 00:00:00 EDT 2017
Highly expressed NRSN2 is related to malignant phenotype in ovarian cancer.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Tang W,Ren A,Xiao H,Sun H,Li B
Sun Jan 01 00:00:00 EST 2017
Highly expressed NRSN2 is related to malignant phenotype in ovarian cancer.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Tang W,Ren A,Xiao H,Sun H,Li B
Sun Jan 01 00:00:00 EST 2017
Growth-induced stress enhances epithelial-mesenchymal transition induced by IL-6 in clear cell renal cell carcinoma via the Akt/GSK-3β/β-catenin signaling pathway.
Oncogenesis
Chen Q,Yang D,Zong H,Zhu L,Wang L,Wang X,Zhu X,Song X,Wang J
Mon Aug 28 00:00:00 EDT 2017
Growth-induced stress enhances epithelial-mesenchymal transition induced by IL-6 in clear cell renal cell carcinoma via the Akt/GSK-3β/β-catenin signaling pathway.
Oncogenesis
Chen Q,Yang D,Zong H,Zhu L,Wang L,Wang X,Zhu X,Song X,Wang J
Mon Aug 28 00:00:00 EDT 2017
Curcumin Suppresses Lung Cancer Stem Cells via Inhibiting Wnt/β-catenin and Sonic Hedgehog Pathways.
Phytotherapy research : PTR
Zhu JY,Yang X,Chen Y,Jiang Y,Wang SJ,Li Y,Wang XQ,Meng Y,Zhu MM,Ma X,Huang C,Wu R,Xie CF,Li XT,Geng SS,Wu JS,Zhong CY,Han HY
Sat Apr 01 00:00:00 EDT 2017
Curcumin Suppresses Lung Cancer Stem Cells via Inhibiting Wnt/β-catenin and Sonic Hedgehog Pathways.
Phytotherapy research : PTR
Zhu JY,Yang X,Chen Y,Jiang Y,Wang SJ,Li Y,Wang XQ,Meng Y,Zhu MM,Ma X,Huang C,Wu R,Xie CF,Li XT,Geng SS,Wu JS,Zhong CY,Han HY
Sat Apr 01 00:00:00 EDT 2017
Downregulation of the Repressor Element 1-Silencing Transcription Factor (REST) Is Associated with Akt-mTOR and Wnt-β-Catenin Signaling in Prion Diseases Models.
Frontiers in molecular neuroscience
Song Z,Shah SZA,Yang W,Dong H,Yang L,Zhou X,Zhao D
Wed Sep 30 00:00:00 EDT 2020
Downregulation of the Repressor Element 1-Silencing Transcription Factor (REST) Is Associated with Akt-mTOR and Wnt-β-Catenin Signaling in Prion Diseases Models.
Frontiers in molecular neuroscience
Song Z,Shah SZA,Yang W,Dong H,Yang L,Zhou X,Zhao D
Wed Sep 30 00:00:00 EDT 2020
Downregulation of the Repressor Element 1-Silencing Transcription Factor (REST) Is Associated with Akt-mTOR and Wnt-β-Catenin Signaling in Prion Diseases Models.
Frontiers in molecular neuroscience
Song Z,Shah SZA,Yang W,Dong H,Yang L,Zhou X,Zhao D
Wed Sep 30 00:00:00 EDT 2020
Downregulation of the Repressor Element 1-Silencing Transcription Factor (REST) Is Associated with Akt-mTOR and Wnt-β-Catenin Signaling in Prion Diseases Models.
Frontiers in molecular neuroscience
Song Z,Shah SZA,Yang W,Dong H,Yang L,Zhou X,Zhao D
Wed Sep 30 00:00:00 EDT 2020
Relaxin attenuates aristolochic acid induced human tubular epithelial cell apoptosis in vitro by activation of the PI3K/Akt signaling pathway.
Apoptosis : an international journal on programmed cell death
Xie XC,Zhao N,Xu QH,Yang X,Xia WK,Chen Q,Wang M,Fei X
Thu Jun 01 00:00:00 EDT 2017
Relaxin attenuates aristolochic acid induced human tubular epithelial cell apoptosis in vitro by activation of the PI3K/Akt signaling pathway.
Apoptosis : an international journal on programmed cell death
Xie XC,Zhao N,Xu QH,Yang X,Xia WK,Chen Q,Wang M,Fei X
Thu Jun 01 00:00:00 EDT 2017
Ku70 Senses HTLV-1 DNA and Modulates HTLV-1 Replication.
Journal of immunology (Baltimore, Md. : 1950)
Wang J,Kang L,Song D,Liu L,Yang S,Ma L,Guo Z,Ding H,Wang H,Yang B
Sun Oct 01 00:00:00 EDT 2017
Ku70 Senses HTLV-1 DNA and Modulates HTLV-1 Replication.
Journal of immunology (Baltimore, Md. : 1950)
Wang J,Kang L,Song D,Liu L,Yang S,Ma L,Guo Z,Ding H,Wang H,Yang B
Sun Oct 01 00:00:00 EDT 2017
Lithium alleviates neurotoxic prion peptide-induced synaptic damage and neuronal death partially by the upregulation of nuclear target REST and the restoration of Wnt signaling.
Neuropharmacology
Song Z,Yang W,Zhou X,Yang L,Zhao D
Fri Sep 01 00:00:00 EDT 2017
Lithium alleviates neurotoxic prion peptide-induced synaptic damage and neuronal death partially by the upregulation of nuclear target REST and the restoration of Wnt signaling.
Neuropharmacology
Song Z,Yang W,Zhou X,Yang L,Zhao D
Fri Sep 01 00:00:00 EDT 2017
Effect of Hypoxia on the Differentiation and the Self-Renewal of Metanephrogenic Mesenchymal Stem Cells.
Stem cells international
Liu S,Song N,He J,Yu X,Guo J,Jiao X,Ding X,Teng J
Wed Sep 30 00:00:00 EDT 2020
Effect of Hypoxia on the Differentiation and the Self-Renewal of Metanephrogenic Mesenchymal Stem Cells.
Stem cells international
Liu S,Song N,He J,Yu X,Guo J,Jiao X,Ding X,Teng J
Wed Sep 30 00:00:00 EDT 2020
Effect of Hypoxia on the Differentiation and the Self-Renewal of Metanephrogenic Mesenchymal Stem Cells.
Stem cells international
Liu S,Song N,He J,Yu X,Guo J,Jiao X,Ding X,Teng J
Wed Sep 30 00:00:00 EDT 2020
Effect of Hypoxia on the Differentiation and the Self-Renewal of Metanephrogenic Mesenchymal Stem Cells.
Stem cells international
Liu S,Song N,He J,Yu X,Guo J,Jiao X,Ding X,Teng J
Wed Sep 30 00:00:00 EDT 2020
The role of the chemokine receptor XCR1 in breast cancer cells.
Breast cancer (Dove Medical Press)
Yang XL,Qi LG,Lin FJ,Ou ZL
Wed Sep 30 00:00:00 EDT 2020
The role of the chemokine receptor XCR1 in breast cancer cells.
Breast cancer (Dove Medical Press)
Yang XL,Qi LG,Lin FJ,Ou ZL
Wed Sep 30 00:00:00 EDT 2020
High glucose impaired estrogen receptor alpha signaling via β-catenin in osteoblastic MC3T3-E1.
The Journal of steroid biochemistry and molecular biology
Wang R,Gao D,Zhou Y,Chen L,Luo B,Yu Y,Li H,Hu J,Huang Q,He M,Peng W,Luo D
Wed Nov 01 00:00:00 EDT 2017
High glucose impaired estrogen receptor alpha signaling via β-catenin in osteoblastic MC3T3-E1.
The Journal of steroid biochemistry and molecular biology
Wang R,Gao D,Zhou Y,Chen L,Luo B,Yu Y,Li H,Hu J,Huang Q,He M,Peng W,Luo D
Wed Nov 01 00:00:00 EDT 2017
High glucose impaired estrogen receptor alpha signaling via β-catenin in osteoblastic MC3T3-E1.
The Journal of steroid biochemistry and molecular biology
Wang R,Gao D,Zhou Y,Chen L,Luo B,Yu Y,Li H,Hu J,Huang Q,He M,Peng W,Luo D
Wed Nov 01 00:00:00 EDT 2017
miR-148a and miR-17-5p synergistically regulate milk TAG synthesis via PPARGC1A and PPARA in goat mammary epithelial cells.
RNA biology
Chen Z,Luo J,Sun S,Cao D,Shi H,Loor JJ
Sat Mar 04 00:00:00 EST 2017
miR-148a and miR-17-5p synergistically regulate milk TAG synthesis via PPARGC1A and PPARA in goat mammary epithelial cells.
RNA biology
Chen Z,Luo J,Sun S,Cao D,Shi H,Loor JJ
Sat Mar 04 00:00:00 EST 2017
KLK4 silencing inhibits the growth of oral squamous cell carcinoma through Wnt/β-catenin signaling pathway.
Cell biology international
Cui Z,Cui Y,Yang S,Luo G,Wang Y,Lou Y,Sun X
Sat Apr 01 00:00:00 EDT 2017
KLK4 silencing inhibits the growth of oral squamous cell carcinoma through Wnt/β-catenin signaling pathway.
Cell biology international
Cui Z,Cui Y,Yang S,Luo G,Wang Y,Lou Y,Sun X
Sat Apr 01 00:00:00 EDT 2017
Spiroplasma eriocheiris induces mouse 3T6-Swiss albino cell apoptosis that associated with the infection mechanism.
Molecular immunology
Hou L,Gu W,Zhu H,Yao W,Wang W,Meng Q
Wed Nov 01 00:00:00 EDT 2017
Spiroplasma eriocheiris induces mouse 3T6-Swiss albino cell apoptosis that associated with the infection mechanism.
Molecular immunology
Hou L,Gu W,Zhu H,Yao W,Wang W,Meng Q
Wed Nov 01 00:00:00 EDT 2017
Osthole inhibits the tumorigenesis of hepatocellular carcinoma cells.
Oncology reports
Lin ZK,Liu J,Jiang GQ,Tan G,Gong P,Luo HF,Li HM,Du J,Ning Z,Xin Y,Wang ZY
Wed Mar 01 00:00:00 EST 2017
Osthole inhibits the tumorigenesis of hepatocellular carcinoma cells.
Oncology reports
Lin ZK,Liu J,Jiang GQ,Tan G,Gong P,Luo HF,Li HM,Du J,Ning Z,Xin Y,Wang ZY
Wed Mar 01 00:00:00 EST 2017
miR-1247-5p functions as a tumor suppressor in human hepatocellular carcinoma by targeting Wnt3.
Oncology reports
Chu Y,Fan W,Guo W,Zhang Y,Wang L,Guo L,Duan X,Wei J,Xu G
Sat Jul 01 00:00:00 EDT 2017
miR-1247-5p functions as a tumor suppressor in human hepatocellular carcinoma by targeting Wnt3.
Oncology reports
Chu Y,Fan W,Guo W,Zhang Y,Wang L,Guo L,Duan X,Wei J,Xu G
Sat Jul 01 00:00:00 EDT 2017
GRHL2 suppresses tumor metastasis via regulation of transcriptional activity of RhoG in non-small cell lung cancer.
American journal of translational research
Pan X,Zhang R,Xie C,Gan M,Yao S,Yao Y,Jin J,Han T,Huang Y,Gong Y,Wang J,Yu B
Wed Sep 30 00:00:00 EDT 2020
GRHL2 suppresses tumor metastasis via regulation of transcriptional activity of RhoG in non-small cell lung cancer.
American journal of translational research
Pan X,Zhang R,Xie C,Gan M,Yao S,Yao Y,Jin J,Han T,Huang Y,Gong Y,Wang J,Yu B
Wed Sep 30 00:00:00 EDT 2020
CO-releasing molecules-2 attenuates ox-LDL-induced injury in HUVECs by ameliorating mitochondrial function and inhibiting Wnt/β-catenin pathway.
Biochemical and biophysical research communications
Sun HJ,Xu DY,Sun YX,Xue T,Zhang CX,Zhang ZX,Lin W,Li KX
Sat Aug 26 00:00:00 EDT 2017
CO-releasing molecules-2 attenuates ox-LDL-induced injury in HUVECs by ameliorating mitochondrial function and inhibiting Wnt/β-catenin pathway.
Biochemical and biophysical research communications
Sun HJ,Xu DY,Sun YX,Xue T,Zhang CX,Zhang ZX,Lin W,Li KX
Sat Aug 26 00:00:00 EDT 2017
Wnt/β-catenin pathway mediates (-)-Epigallocatechin-3-gallate (EGCG) inhibition of lung cancer stem cells.
Biochemical and biophysical research communications
Zhu J,Jiang Y,Yang X,Wang S,Xie C,Li X,Li Y,Chen Y,Wang X,Meng Y,Zhu M,Wu R,Huang C,Ma X,Geng S,Wu J,Zhong C
Sun Jan 01 00:00:00 EST 2017
Wnt/β-catenin pathway mediates (-)-Epigallocatechin-3-gallate (EGCG) inhibition of lung cancer stem cells.
Biochemical and biophysical research communications
Zhu J,Jiang Y,Yang X,Wang S,Xie C,Li X,Li Y,Chen Y,Wang X,Meng Y,Zhu M,Wu R,Huang C,Ma X,Geng S,Wu J,Zhong C
Sun Jan 01 00:00:00 EST 2017
Relaxin Attenuates Contrast-Induced Human Proximal Tubular Epithelial Cell Apoptosis by Activation of the PI3K/Akt Signaling Pathway In Vitro.
BioMed research international
Xie XC,Cao Y,Yang X,Xu QH,Wei W,Wang M
Tue Feb 27 00:00:00 EST 2018
Relaxin Attenuates Contrast-Induced Human Proximal Tubular Epithelial Cell Apoptosis by Activation of the PI3K/Akt Signaling Pathway In Vitro.
BioMed research international
Xie XC,Cao Y,Yang X,Xu QH,Wei W,Wang M
Tue Feb 27 00:00:00 EST 2018
β-catenin coordinates with Jup and the TCF1/GATA6 axis to regulate human embryonic stem cell fate.
Developmental biology
Sun H,Wang X,Liu K,Guo M,Zhang Y,Ying QL,Ye S
Wed Nov 15 00:00:00 EST 2017
β-catenin coordinates with Jup and the TCF1/GATA6 axis to regulate human embryonic stem cell fate.
Developmental biology
Sun H,Wang X,Liu K,Guo M,Zhang Y,Ying QL,Ye S
Wed Nov 15 00:00:00 EST 2017
iTRAQ-coupled 2D LC/MS-MS analysis of CXCR7-transfected papillary thyroid carcinoma cells: A new insight into CXCR7 regulation of papillary thyroid carcinoma progression and identification of potential biomarkers.
Oncology letters
Zhang H,Yang L,Liu Z,Liu C,Teng X,Zhang L,Yin B,Liu Z
Fri Sep 01 00:00:00 EDT 2017
iTRAQ-coupled 2D LC/MS-MS analysis of CXCR7-transfected papillary thyroid carcinoma cells: A new insight into CXCR7 regulation of papillary thyroid carcinoma progression and identification of potential biomarkers.
Oncology letters
Zhang H,Yang L,Liu Z,Liu C,Teng X,Zhang L,Yin B,Liu Z
Fri Sep 01 00:00:00 EDT 2017
Sulfiredoxin May Promote Cervical Cancer Metastasis via Wnt/β-Catenin Signaling Pathway.
International journal of molecular sciences
Lan K,Zhao Y,Fan Y,Ma B,Yang S,Liu Q,Linghu H,Wang H
Thu Apr 27 00:00:00 EDT 2017
Sulfiredoxin May Promote Cervical Cancer Metastasis via Wnt/β-Catenin Signaling Pathway.
International journal of molecular sciences
Lan K,Zhao Y,Fan Y,Ma B,Yang S,Liu Q,Linghu H,Wang H
Thu Apr 27 00:00:00 EDT 2017
Resveratrol reverses Doxorubicin resistance by inhibiting epithelial-mesenchymal transition (EMT) through modulating PTEN/Akt signaling pathway in gastric cancer.
Journal of experimental & clinical cancer research : CR
Xu J,Liu D,Niu H,Zhu G,Xu Y,Ye D,Li J,Zhang Q
Thu Jan 26 00:00:00 EST 2017
Resveratrol reverses Doxorubicin resistance by inhibiting epithelial-mesenchymal transition (EMT) through modulating PTEN/Akt signaling pathway in gastric cancer.
Journal of experimental & clinical cancer research : CR
Xu J,Liu D,Niu H,Zhu G,Xu Y,Ye D,Li J,Zhang Q
Thu Jan 26 00:00:00 EST 2017
Resveratrol reverses Doxorubicin resistance by inhibiting epithelial-mesenchymal transition (EMT) through modulating PTEN/Akt signaling pathway in gastric cancer.
Journal of experimental & clinical cancer research : CR
Xu J,Liu D,Niu H,Zhu G,Xu Y,Ye D,Li J,Zhang Q
Thu Jan 26 00:00:00 EST 2017
Resveratrol reverses Doxorubicin resistance by inhibiting epithelial-mesenchymal transition (EMT) through modulating PTEN/Akt signaling pathway in gastric cancer.
Journal of experimental & clinical cancer research : CR
Xu J,Liu D,Niu H,Zhu G,Xu Y,Ye D,Li J,Zhang Q
Thu Jan 26 00:00:00 EST 2017
(-)-Epigallocatechin-3-Gallate Inhibits Colorectal Cancer Stem Cells by Suppressing Wnt/β-Catenin Pathway.
Nutrients
Chen Y,Wang XQ,Zhang Q,Zhu JY,Li Y,Xie CF,Li XT,Wu JS,Geng SS,Zhong CY,Han HY
Sat Jun 03 00:00:00 EDT 2017
(-)-Epigallocatechin-3-Gallate Inhibits Colorectal Cancer Stem Cells by Suppressing Wnt/β-Catenin Pathway.
Nutrients
Chen Y,Wang XQ,Zhang Q,Zhu JY,Li Y,Xie CF,Li XT,Wu JS,Geng SS,Zhong CY,Han HY
Sat Jun 03 00:00:00 EDT 2017
Transcriptional suppression of microRNA-27a contributes to laryngeal cancer differentiation via GSK-3β-involved Wnt/β-catenin pathway.
Oncotarget
Chen S,Sun YY,Zhang ZX,Li YH,Xu ZM,Fu WN
Tue Feb 28 00:00:00 EST 2017
Transcriptional suppression of microRNA-27a contributes to laryngeal cancer differentiation via GSK-3β-involved Wnt/β-catenin pathway.
Oncotarget
Chen S,Sun YY,Zhang ZX,Li YH,Xu ZM,Fu WN
Tue Feb 28 00:00:00 EST 2017
Hypoxia-induced PLOD2 promotes proliferation, migration and invasion via PI3K/Akt signaling in glioma.
Oncotarget
Song Y,Zheng S,Wang J,Long H,Fang L,Wang G,Li Z,Que T,Liu Y,Li Y,Zhang X,Fang W,Qi S
Tue Jun 27 00:00:00 EDT 2017
Hypoxia-induced PLOD2 promotes proliferation, migration and invasion via PI3K/Akt signaling in glioma.
Oncotarget
Song Y,Zheng S,Wang J,Long H,Fang L,Wang G,Li Z,Que T,Liu Y,Li Y,Zhang X,Fang W,Qi S
Tue Jun 27 00:00:00 EDT 2017
TLR4 Influences Hepatitis B Virus Related Hepatocellular Carcinoma by Regulating the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Yin Y,Li F,Li S,Cai J,Shi J,Jiang Y
Fri Aug 04 00:00:00 EDT 2017
TLR4 Influences Hepatitis B Virus Related Hepatocellular Carcinoma by Regulating the Wnt/β-Catenin Pathway.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Yin Y,Li F,Li S,Cai J,Shi J,Jiang Y
Fri Aug 04 00:00:00 EDT 2017
TIMELESS confers cisplatin resistance in nasopharyngeal carcinoma by activating the Wnt/β-catenin signaling pathway and promoting the epithelial mesenchymal transition.
Cancer letters
Liu SL,Lin HX,Lin CY,Sun XQ,Ye LP,Qiu F,Wen W,Hua X,Wu XQ,Li J,Song LB,Guo L
Mon Aug 28 00:00:00 EDT 2017
Calreticulin promotes EGF-induced EMT in pancreatic cancer cells via Integrin/EGFR-ERK/MAPK signaling pathway.
Cell death & disease
Sheng W,Chen C,Dong M,Wang G,Zhou J,Song H,Li Y,Zhang J,Ding S
Thu Oct 26 00:00:00 EDT 2017
Calreticulin promotes EGF-induced EMT in pancreatic cancer cells via Integrin/EGFR-ERK/MAPK signaling pathway.
Cell death & disease
Sheng W,Chen C,Dong M,Wang G,Zhou J,Song H,Li Y,Zhang J,Ding S
Thu Oct 26 00:00:00 EDT 2017
MTA1 promotes epithelial to mesenchymal transition and metastasis in non-small-cell lung cancer.
Oncotarget
Ma K,Fan Y,Dong X,Dong D,Guo Y,Wei X,Ning J,Geng Q,Wang C,Hu Y,Li M,Niu W,Li E,Wu Y
Tue Jun 13 00:00:00 EDT 2017
c-Met, CREB1 and EGFR are involved in miR-493-5p inhibition of EMT via AKT/GSK-3β/Snail signaling in prostate cancer.
Oncotarget
Wang S,Wang X,Li J,Meng S,Liang Z,Xu X,Zhu Y,Li S,Wu J,Xu M,Ji A,Lin Y,Liu B,Zheng X,Xie B,Xie L
Tue Oct 10 00:00:00 EDT 2017
iTRAQ-based Proteomic Analysis of Porcine Kidney Epithelial PK15 cells Infected with Pseudorabies virus.
Scientific reports
Yang S,Pei Y,Zhao A
Tue Apr 04 00:00:00 EDT 2017
iTRAQ-based Proteomic Analysis of Porcine Kidney Epithelial PK15 cells Infected with Pseudorabies virus.
Scientific reports
Yang S,Pei Y,Zhao A
Tue Apr 04 00:00:00 EDT 2017
Hypoxia and TGF-β1 induced PLOD2 expression improve the migration and invasion of cervical cancer cells by promoting epithelial-to-mesenchymal transition (EMT) and focal adhesion formation.
Cancer cell international
Xu F,Zhang J,Hu G,Liu L,Liang W
Wed Sep 30 00:00:00 EDT 2020
Hypoxia and TGF-β1 induced PLOD2 expression improve the migration and invasion of cervical cancer cells by promoting epithelial-to-mesenchymal transition (EMT) and focal adhesion formation.
Cancer cell international
Xu F,Zhang J,Hu G,Liu L,Liang W
Wed Sep 30 00:00:00 EDT 2020
Hypoxia and TGF-β1 induced PLOD2 expression improve the migration and invasion of cervical cancer cells by promoting epithelial-to-mesenchymal transition (EMT) and focal adhesion formation.
Cancer cell international
Xu F,Zhang J,Hu G,Liu L,Liang W
Wed Sep 30 00:00:00 EDT 2020
Hypoxia and TGF-β1 induced PLOD2 expression improve the migration and invasion of cervical cancer cells by promoting epithelial-to-mesenchymal transition (EMT) and focal adhesion formation.
Cancer cell international
Xu F,Zhang J,Hu G,Liu L,Liang W
Wed Sep 30 00:00:00 EDT 2020
Apobec-1 complementation factor regulates cell migration and apoptosis through Dickkopf1 by acting on its 3' untranslated region in MCF7 cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Yan X,Li Q,Ni D,Xie Y,He Q,Wan Q,Liu Y,Lyu Z,Mao Z,Zhou Q
Thu Jun 01 00:00:00 EDT 2017
Apobec-1 complementation factor regulates cell migration and apoptosis through Dickkopf1 by acting on its 3' untranslated region in MCF7 cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Yan X,Li Q,Ni D,Xie Y,He Q,Wan Q,Liu Y,Lyu Z,Mao Z,Zhou Q
Thu Jun 01 00:00:00 EDT 2017
Long noncoding RNA DANCR increases stemness features of hepatocellular carcinoma by derepression of CTNNB1.
Hepatology (Baltimore, Md.)
Yuan SX,Wang J,Yang F,Tao QF,Zhang J,Wang LL,Yang Y,Liu H,Wang ZG,Xu QG,Fan J,Liu L,Sun SH,Zhou WP
Mon Feb 01 00:00:00 EST 2016
Long noncoding RNA DANCR increases stemness features of hepatocellular carcinoma by derepression of CTNNB1.
Hepatology (Baltimore, Md.)
Yuan SX,Wang J,Yang F,Tao QF,Zhang J,Wang LL,Yang Y,Liu H,Wang ZG,Xu QG,Fan J,Liu L,Sun SH,Zhou WP
Mon Feb 01 00:00:00 EST 2016
Disrupting MALAT1/miR-200c sponge decreases invasion and migration in endometrioid endometrial carcinoma.
Cancer letters
Li Q,Zhang C,Chen R,Xiong H,Qiu F,Liu S,Zhang M,Wang F,Wang Y,Zhou X,Xiao G,Wang X,Jiang Q
Thu Dec 01 00:00:00 EST 2016
Disrupting MALAT1/miR-200c sponge decreases invasion and migration in endometrioid endometrial carcinoma.
Cancer letters
Li Q,Zhang C,Chen R,Xiong H,Qiu F,Liu S,Zhang M,Wang F,Wang Y,Zhou X,Xiao G,Wang X,Jiang Q
Thu Dec 01 00:00:00 EST 2016
Disrupting MALAT1/miR-200c sponge decreases invasion and migration in endometrioid endometrial carcinoma.
Cancer letters
Li Q,Zhang C,Chen R,Xiong H,Qiu F,Liu S,Zhang M,Wang F,Wang Y,Zhou X,Xiao G,Wang X,Jiang Q
Thu Dec 01 00:00:00 EST 2016
Disrupting MALAT1/miR-200c sponge decreases invasion and migration in endometrioid endometrial carcinoma.
Cancer letters
Li Q,Zhang C,Chen R,Xiong H,Qiu F,Liu S,Zhang M,Wang F,Wang Y,Zhou X,Xiao G,Wang X,Jiang Q
Thu Dec 01 00:00:00 EST 2016
Altered Wnt Signaling Pathway in Cognitive Impairment Caused by Chronic Intermittent Hypoxia: Focus on Glycogen Synthase Kinase-3β and β-catenin.
Chinese medical journal
Pan YY,Deng Y,Xie S,Wang ZH,Wang Y,Ren J,Liu HG
Tue Apr 05 00:00:00 EDT 2016
Altered Wnt Signaling Pathway in Cognitive Impairment Caused by Chronic Intermittent Hypoxia: Focus on Glycogen Synthase Kinase-3β and β-catenin.
Chinese medical journal
Pan YY,Deng Y,Xie S,Wang ZH,Wang Y,Ren J,Liu HG
Tue Apr 05 00:00:00 EDT 2016
Magnetic gold nanoparticle-mediated small interference RNA silencing Bag-1 gene for colon cancer therapy.
Oncology reports
Huang W,Liu Z,Zhou G,Tian A,Sun N
Mon Feb 01 00:00:00 EST 2016
Magnetic gold nanoparticle-mediated small interference RNA silencing Bag-1 gene for colon cancer therapy.
Oncology reports
Huang W,Liu Z,Zhou G,Tian A,Sun N
Mon Feb 01 00:00:00 EST 2016
Perichondrium mesenchymal stem cells inhibit the growth of breast cancer cells via the DKK-1/Wnt/β-catenin signaling pathway.
Oncology reports
Li M,Cai H,Yang Y,Zhang J,Sun K,Yan Y,Qu H,Wang W,Wang J,Duan X
Mon Aug 01 00:00:00 EDT 2016
Perichondrium mesenchymal stem cells inhibit the growth of breast cancer cells via the DKK-1/Wnt/β-catenin signaling pathway.
Oncology reports
Li M,Cai H,Yang Y,Zhang J,Sun K,Yan Y,Qu H,Wang W,Wang J,Duan X
Mon Aug 01 00:00:00 EDT 2016
Exosomes derived from mineralizing osteoblasts promote ST2 cell osteogenic differentiation by alteration of microRNA expression.
FEBS letters
Cui Y,Luan J,Li H,Zhou X,Han J
Fri Jan 01 00:00:00 EST 2016
Exosomes derived from mineralizing osteoblasts promote ST2 cell osteogenic differentiation by alteration of microRNA expression.
FEBS letters
Cui Y,Luan J,Li H,Zhou X,Han J
Fri Jan 01 00:00:00 EST 2016
High expression of hexokinase domain containing 1 is associated with poor prognosis and aggressive phenotype in hepatocarcinoma.
Biochemical and biophysical research communications
Zhang Z,Huang S,Wang H,Wu J,Chen D,Peng B,Zhou Q
Fri Jun 10 00:00:00 EDT 2016
High expression of hexokinase domain containing 1 is associated with poor prognosis and aggressive phenotype in hepatocarcinoma.
Biochemical and biophysical research communications
Zhang Z,Huang S,Wang H,Wu J,Chen D,Peng B,Zhou Q
Fri Jun 10 00:00:00 EDT 2016
Identification of novel NRAGE involved in the radioresistance of esophageal cancer cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Zhou H,Zhang G,Xue X,Yang Y,Yang Y,Chang X,Ge X,Xiao Z,Guo H,Wang Y
Fri Jul 01 00:00:00 EDT 2016
Identification of novel NRAGE involved in the radioresistance of esophageal cancer cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Zhou H,Zhang G,Xue X,Yang Y,Yang Y,Chang X,Ge X,Xiao Z,Guo H,Wang Y
Fri Jul 01 00:00:00 EDT 2016
Identification of novel NRAGE involved in the radioresistance of esophageal cancer cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Zhou H,Zhang G,Xue X,Yang Y,Yang Y,Chang X,Ge X,Xiao Z,Guo H,Wang Y
Fri Jul 01 00:00:00 EDT 2016
Identification of novel NRAGE involved in the radioresistance of esophageal cancer cells.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Zhou H,Zhang G,Xue X,Yang Y,Yang Y,Chang X,Ge X,Xiao Z,Guo H,Wang Y
Fri Jul 01 00:00:00 EDT 2016
Downregulation of HP1α suppresses proliferation of cholangiocarcinoma by restoring SFRP1 expression.
Oncotarget
Cheng W,Tian L,Wang B,Qi Y,Huang W,Li H,Chen YJ
Tue Jul 26 00:00:00 EDT 2016
Downregulation of HP1α suppresses proliferation of cholangiocarcinoma by restoring SFRP1 expression.
Oncotarget
Cheng W,Tian L,Wang B,Qi Y,Huang W,Li H,Chen YJ
Tue Jul 26 00:00:00 EDT 2016
MicroRNA-655-3p functions as a tumor suppressor by regulating ADAM10 and β-catenin pathway in Hepatocellular Carcinoma.
Journal of experimental & clinical cancer research : CR
Wu G,Zheng K,Xia S,Wang Y,Meng X,Qin X,Cheng Y
Sat Jun 04 00:00:00 EDT 2016
MicroRNA-655-3p functions as a tumor suppressor by regulating ADAM10 and β-catenin pathway in Hepatocellular Carcinoma.
Journal of experimental & clinical cancer research : CR
Wu G,Zheng K,Xia S,Wang Y,Meng X,Qin X,Cheng Y
Sat Jun 04 00:00:00 EDT 2016
High Expression of XRCC6 Promotes Human Osteosarcoma Cell Proliferation through the β-Catenin/Wnt Signaling Pathway and Is Associated with Poor Prognosis.
International journal of molecular sciences
Zhu B,Cheng D,Li S,Zhou S,Yang Q
Fri Jul 22 00:00:00 EDT 2016
High Expression of XRCC6 Promotes Human Osteosarcoma Cell Proliferation through the β-Catenin/Wnt Signaling Pathway and Is Associated with Poor Prognosis.
International journal of molecular sciences
Zhu B,Cheng D,Li S,Zhou S,Yang Q
Fri Jul 22 00:00:00 EDT 2016
REST alleviates neurotoxic prion peptide-induced synaptic abnormalities, neurofibrillary degeneration and neuronal death partially via LRP6-mediated Wnt-β-catenin signaling.
Oncotarget
Song Z,Zhu T,Zhou X,Barrow P,Yang W,Cui Y,Yang L,Zhao D
Tue Mar 15 00:00:00 EDT 2016
REST alleviates neurotoxic prion peptide-induced synaptic abnormalities, neurofibrillary degeneration and neuronal death partially via LRP6-mediated Wnt-β-catenin signaling.
Oncotarget
Song Z,Zhu T,Zhou X,Barrow P,Yang W,Cui Y,Yang L,Zhao D
Tue Mar 15 00:00:00 EDT 2016
REST alleviates neurotoxic prion peptide-induced synaptic abnormalities, neurofibrillary degeneration and neuronal death partially via LRP6-mediated Wnt-β-catenin signaling.
Oncotarget
Song Z,Zhu T,Zhou X,Barrow P,Yang W,Cui Y,Yang L,Zhao D
Tue Mar 15 00:00:00 EDT 2016
REST alleviates neurotoxic prion peptide-induced synaptic abnormalities, neurofibrillary degeneration and neuronal death partially via LRP6-mediated Wnt-β-catenin signaling.
Oncotarget
Song Z,Zhu T,Zhou X,Barrow P,Yang W,Cui Y,Yang L,Zhao D
Tue Mar 15 00:00:00 EDT 2016
PDIA6 promotes the proliferation of HeLa cells through activating the Wnt/β-catenin signaling pathway.
Oncotarget
Gao H,Sun B,Fu H,Chi X,Wang F,Qi X,Hu J,Shao S
Tue Aug 16 00:00:00 EDT 2016
PDIA6 promotes the proliferation of HeLa cells through activating the Wnt/β-catenin signaling pathway.
Oncotarget
Gao H,Sun B,Fu H,Chi X,Wang F,Qi X,Hu J,Shao S
Tue Aug 16 00:00:00 EDT 2016
PDIA6 promotes the proliferation of HeLa cells through activating the Wnt/β-catenin signaling pathway.
Oncotarget
Gao H,Sun B,Fu H,Chi X,Wang F,Qi X,Hu J,Shao S
Tue Aug 16 00:00:00 EDT 2016
PDIA6 promotes the proliferation of HeLa cells through activating the Wnt/β-catenin signaling pathway.
Oncotarget
Gao H,Sun B,Fu H,Chi X,Wang F,Qi X,Hu J,Shao S
Tue Aug 16 00:00:00 EDT 2016
Suppression of prostate cancer progression by cancer cell stemness inhibitor napabucasin.
Cancer medicine
Zhang Y,Jin Z,Zhou H,Ou X,Xu Y,Li H,Liu C,Li B
Wed Jun 01 00:00:00 EDT 2016
Evodiamine exerts anti-tumor effects against hepatocellular carcinoma through inhibiting β-catenin-mediated angiogenesis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Shi L,Yang F,Luo F,Liu Y,Zhang F,Zou M,Liu Q
Thu Sep 01 00:00:00 EDT 2016
Evodiamine exerts anti-tumor effects against hepatocellular carcinoma through inhibiting β-catenin-mediated angiogenesis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Shi L,Yang F,Luo F,Liu Y,Zhang F,Zou M,Liu Q
Thu Sep 01 00:00:00 EDT 2016
Evodiamine exerts anti-tumor effects against hepatocellular carcinoma through inhibiting β-catenin-mediated angiogenesis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Shi L,Yang F,Luo F,Liu Y,Zhang F,Zou M,Liu Q
Thu Sep 01 00:00:00 EDT 2016
Evodiamine exerts anti-tumor effects against hepatocellular carcinoma through inhibiting β-catenin-mediated angiogenesis.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Shi L,Yang F,Luo F,Liu Y,Zhang F,Zou M,Liu Q
Thu Sep 01 00:00:00 EDT 2016
Persisting and Increasing Neutrophil Infiltration Associates with Gastric Carcinogenesis and E-cadherin Downregulation.
Scientific reports
Fu H,Ma Y,Yang M,Zhang C,Huang H,Xia Y,Lu L,Jin W,Cui D
Thu Jul 14 00:00:00 EDT 2016
Persisting and Increasing Neutrophil Infiltration Associates with Gastric Carcinogenesis and E-cadherin Downregulation.
Scientific reports
Fu H,Ma Y,Yang M,Zhang C,Huang H,Xia Y,Lu L,Jin W,Cui D
Thu Jul 14 00:00:00 EDT 2016
Paper-based microreactor array for rapid screening of cell signaling cascades.
Lab on a chip
Huang CH,Lei KF,Tsang NM
Sun Aug 07 00:00:00 EDT 2016
Paper-based microreactor array for rapid screening of cell signaling cascades.
Lab on a chip
Huang CH,Lei KF,Tsang NM
Sun Aug 07 00:00:00 EDT 2016
Silencing Bag-1 gene via magnetic gold nanoparticle-delivered siRNA plasmid for colorectal cancer therapy in vivo and in vitro.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Huang W,Liu Z,Zhou G,Ling J,Tian A,Sun N
Mon Aug 01 00:00:00 EDT 2016
Silencing Bag-1 gene via magnetic gold nanoparticle-delivered siRNA plasmid for colorectal cancer therapy in vivo and in vitro.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Huang W,Liu Z,Zhou G,Ling J,Tian A,Sun N
Mon Aug 01 00:00:00 EDT 2016
Silencing Bag-1 gene via magnetic gold nanoparticle-delivered siRNA plasmid for colorectal cancer therapy in vivo and in vitro.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Huang W,Liu Z,Zhou G,Ling J,Tian A,Sun N
Mon Aug 01 00:00:00 EDT 2016
Silencing Bag-1 gene via magnetic gold nanoparticle-delivered siRNA plasmid for colorectal cancer therapy in vivo and in vitro.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Huang W,Liu Z,Zhou G,Ling J,Tian A,Sun N
Mon Aug 01 00:00:00 EDT 2016
Restoring Retinoic Acid Attenuates Intestinal Inflammation and Tumorigenesis in APCMin/+ Mice.
Cancer immunology research
Penny HL,Prestwood TR,Bhattacharya N,Sun F,Kenkel JA,Davidson MG,Shen L,Zuniga LA,Seeley ES,Pai R,Choi O,Tolentino L,Wang J,Napoli JL,Engleman EG
Tue Nov 01 00:00:00 EDT 2016
YPEL3 suppresses epithelial-mesenchymal transition and metastasis of nasopharyngeal carcinoma cells through the Wnt/β-catenin signaling pathway.
Journal of experimental & clinical cancer research : CR
Zhang J,Wen X,Ren XY,Li YQ,Tang XR,Wang YQ,He QM,Yang XJ,Sun Y,Liu N,Ma J
Mon Jul 11 00:00:00 EDT 2016
YPEL3 suppresses epithelial-mesenchymal transition and metastasis of nasopharyngeal carcinoma cells through the Wnt/β-catenin signaling pathway.
Journal of experimental & clinical cancer research : CR
Zhang J,Wen X,Ren XY,Li YQ,Tang XR,Wang YQ,He QM,Yang XJ,Sun Y,Liu N,Ma J
Mon Jul 11 00:00:00 EDT 2016
IL-27 inhibits the TGF-β1-induced epithelial-mesenchymal transition in alveolar epithelial cells.
BMC cell biology
Dong Z,Tai W,Lei W,Wang Y,Li Z,Zhang T
Tue Mar 01 00:00:00 EST 2016
IL-27 inhibits the TGF-β1-induced epithelial-mesenchymal transition in alveolar epithelial cells.
BMC cell biology
Dong Z,Tai W,Lei W,Wang Y,Li Z,Zhang T
Tue Mar 01 00:00:00 EST 2016
MicroRNA-421 regulated by HIF-1α promotes metastasis, inhibits apoptosis, and induces cisplatin resistance by targeting E-cadherin and caspase-3 in gastric cancer.
Oncotarget
Ge X,Liu X,Lin F,Li P,Liu K,Geng R,Dai C,Lin Y,Tang W,Wu Z,Chang J,Lu J,Li J
Tue Apr 26 00:00:00 EDT 2016
MicroRNA-421 regulated by HIF-1α promotes metastasis, inhibits apoptosis, and induces cisplatin resistance by targeting E-cadherin and caspase-3 in gastric cancer.
Oncotarget
Ge X,Liu X,Lin F,Li P,Liu K,Geng R,Dai C,Lin Y,Tang W,Wu Z,Chang J,Lu J,Li J
Tue Apr 26 00:00:00 EDT 2016
The Hedgehog signalling pathway mediates drug response of MCF-7 mammosphere cells in breast cancer patients.
Clinical science (London, England : 1979)
He M,Fu Y,Yan Y,Xiao Q,Wu H,Yao W,Zhao H,Zhao L,Jiang Q,Yu Z,Jin F,Mi X,Wang E,Cui Z,Fu L,Chen J,Wei M
Sun Nov 01 00:00:00 EDT 2015
The Hedgehog signalling pathway mediates drug response of MCF-7 mammosphere cells in breast cancer patients.
Clinical science (London, England : 1979)
He M,Fu Y,Yan Y,Xiao Q,Wu H,Yao W,Zhao H,Zhao L,Jiang Q,Yu Z,Jin F,Mi X,Wang E,Cui Z,Fu L,Chen J,Wei M
Sun Nov 01 00:00:00 EDT 2015
Deregulation of secreted frizzled-related proteins is associated with aberrant β-catenin activation in the carcinogenesis of oral submucous fibrosis.
OncoTargets and therapy
Zhou S,Chen L,Mashrah M,Zhu Y,Liu J,Yang X,He Z,Wang L,Xiang T,Yao Z,Guo F,Yang W,Zhang C
Wed Oct 28 00:00:00 EDT 2015
Deregulation of secreted frizzled-related proteins is associated with aberrant β-catenin activation in the carcinogenesis of oral submucous fibrosis.
OncoTargets and therapy
Zhou S,Chen L,Mashrah M,Zhu Y,Liu J,Yang X,He Z,Wang L,Xiang T,Yao Z,Guo F,Yang W,Zhang C
Wed Oct 28 00:00:00 EDT 2015
Irradiated fibroblasts promote epithelial-mesenchymal transition and HDGF expression of esophageal squamous cell carcinoma.
Biochemical and biophysical research communications
Bao CH,Wang XT,Ma W,Wang NN,Un Nesa E,Wang JB,Wang C,Jia YB,Wang K,Tian H,Cheng YF
Fri Mar 06 00:00:00 EST 2015
Irradiated fibroblasts promote epithelial-mesenchymal transition and HDGF expression of esophageal squamous cell carcinoma.
Biochemical and biophysical research communications
Bao CH,Wang XT,Ma W,Wang NN,Un Nesa E,Wang JB,Wang C,Jia YB,Wang K,Tian H,Cheng YF
Fri Mar 06 00:00:00 EST 2015
Irradiated fibroblasts promote epithelial-mesenchymal transition and HDGF expression of esophageal squamous cell carcinoma.
Biochemical and biophysical research communications
Bao CH,Wang XT,Ma W,Wang NN,Un Nesa E,Wang JB,Wang C,Jia YB,Wang K,Tian H,Cheng YF
Fri Mar 06 00:00:00 EST 2015
Irradiated fibroblasts promote epithelial-mesenchymal transition and HDGF expression of esophageal squamous cell carcinoma.
Biochemical and biophysical research communications
Bao CH,Wang XT,Ma W,Wang NN,Un Nesa E,Wang JB,Wang C,Jia YB,Wang K,Tian H,Cheng YF
Fri Mar 06 00:00:00 EST 2015
The Role and Mechanism of α-Klotho in the Calcification of Rat Aortic Vascular Smooth Muscle Cells.
BioMed research international
Chen T,Mao H,Chen C,Wu L,Wang N,Zhao X,Qian J,Xing C
Thu Sep 08 00:00:00 EDT 2016
The Role and Mechanism of α-Klotho in the Calcification of Rat Aortic Vascular Smooth Muscle Cells.
BioMed research international
Chen T,Mao H,Chen C,Wu L,Wang N,Zhao X,Qian J,Xing C
Thu Sep 08 00:00:00 EDT 2016
Huaier Aqueous Extract Induces Hepatocellular Carcinoma Cells Arrest in S Phase via JNK Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Zhang C,Zhang J,Li X,Sun N,Yu R,Zhao B,Yu D,Cheng Y,Liu Y
Mon Aug 03 00:00:00 EDT 2015
Huaier Aqueous Extract Induces Hepatocellular Carcinoma Cells Arrest in S Phase via JNK Signaling Pathway.
Evidence-based complementary and alternative medicine : eCAM
Zhang C,Zhang J,Li X,Sun N,Yu R,Zhao B,Yu D,Cheng Y,Liu Y
Mon Aug 03 00:00:00 EDT 2015
DACH1 is a novel predictive and prognostic biomarker in hepatocellular carcinoma as a negative regulator of Wnt/β-catenin signaling.
Oncotarget
Liu Y,Zhou R,Yuan X,Han N,Zhou S,Xu H,Guo M,Yu S,Zhang C,Yin T,Wu K
Mon Apr 20 00:00:00 EDT 2015
DACH1 is a novel predictive and prognostic biomarker in hepatocellular carcinoma as a negative regulator of Wnt/β-catenin signaling.
Oncotarget
Liu Y,Zhou R,Yuan X,Han N,Zhou S,Xu H,Guo M,Yu S,Zhang C,Yin T,Wu K
Mon Apr 20 00:00:00 EDT 2015
Induction of Apoptosis in Human Leukemic Cell Lines by Diallyl Disulfide via Modulation of EGFR/ERK/PKM2 Signaling Pathways.
Asian Pacific journal of cancer prevention : APJCP
Luo N,Zhao LC,Shi QQ,Feng ZQ,Chen DL,Li J
Tue Jan 26 00:00:00 EST 2016
Induction of Apoptosis in Human Leukemic Cell Lines by Diallyl Disulfide via Modulation of EGFR/ERK/PKM2 Signaling Pathways.
Asian Pacific journal of cancer prevention : APJCP
Luo N,Zhao LC,Shi QQ,Feng ZQ,Chen DL,Li J
Tue Jan 26 00:00:00 EST 2016
MiR-146a-5p suppresses activation and proliferation of hepatic stellate cells in nonalcoholic fibrosing steatohepatitis through directly targeting Wnt1 and Wnt5a.
Scientific reports
Du J,Niu X,Wang Y,Kong L,Wang R,Zhang Y,Zhao S,Nan Y
Thu Nov 05 00:00:00 EST 2015
MiR-146a-5p suppresses activation and proliferation of hepatic stellate cells in nonalcoholic fibrosing steatohepatitis through directly targeting Wnt1 and Wnt5a.
Scientific reports
Du J,Niu X,Wang Y,Kong L,Wang R,Zhang Y,Zhao S,Nan Y
Thu Nov 05 00:00:00 EST 2015
Potent effects of dioscin against liver fibrosis.
Scientific reports
Zhang X,Han X,Yin L,Xu L,Qi Y,Xu Y,Sun H,Lin Y,Liu K,Peng J
Wed Apr 08 00:00:00 EDT 2015
Potent effects of dioscin against liver fibrosis.
Scientific reports
Zhang X,Han X,Yin L,Xu L,Qi Y,Xu Y,Sun H,Lin Y,Liu K,Peng J
Wed Apr 08 00:00:00 EDT 2015
Low SFRP1 Expression Correlates with Poor Prognosis and Promotes Cell Invasion by Activating the Wnt/β-Catenin Signaling Pathway in NPC.
Cancer prevention research (Philadelphia, Pa.)
Ren XY,Zhou GQ,Jiang W,Sun Y,Xu YF,Li YQ,Tang XR,Wen X,He QM,Yang XJ,Liu N,Ma J
Thu Oct 01 00:00:00 EDT 2015
Low SFRP1 Expression Correlates with Poor Prognosis and Promotes Cell Invasion by Activating the Wnt/β-Catenin Signaling Pathway in NPC.
Cancer prevention research (Philadelphia, Pa.)
Ren XY,Zhou GQ,Jiang W,Sun Y,Xu YF,Li YQ,Tang XR,Wen X,He QM,Yang XJ,Liu N,Ma J
Thu Oct 01 00:00:00 EDT 2015
Elevated GAPDH expression is associated with the proliferation and invasion of lung and esophageal squamous cell carcinomas.
Proteomics
Hao L,Zhou X,Liu S,Sun M,Song Y,Du S,Sun B,Guo C,Gong L,Hu J,Guan H,Shao S
Tue Sep 01 00:00:00 EDT 2015
Notch1 promotes hepatitis B virus X protein-induced hepatocarcinogenesis via Wnt/β-catenin pathway.
International journal of oncology
Sun Q,Wang R,Luo J,Wang P,Xiong S,Liu M,Cheng B
Wed Oct 01 00:00:00 EDT 2014
Notch1 promotes hepatitis B virus X protein-induced hepatocarcinogenesis via Wnt/β-catenin pathway.
International journal of oncology
Sun Q,Wang R,Luo J,Wang P,Xiong S,Liu M,Cheng B
Wed Oct 01 00:00:00 EDT 2014
Notch1 promotes hepatitis B virus X protein-induced hepatocarcinogenesis via Wnt/β-catenin pathway.
International journal of oncology
Sun Q,Wang R,Luo J,Wang P,Xiong S,Liu M,Cheng B
Wed Oct 01 00:00:00 EDT 2014
Notch1 promotes hepatitis B virus X protein-induced hepatocarcinogenesis via Wnt/β-catenin pathway.
International journal of oncology
Sun Q,Wang R,Luo J,Wang P,Xiong S,Liu M,Cheng B
Wed Oct 01 00:00:00 EDT 2014
Notch1 promotes hepatitis B virus X protein-induced hepatocarcinogenesis via Wnt/β-catenin pathway.
International journal of oncology
Sun Q,Wang R,Luo J,Wang P,Xiong S,Liu M,Cheng B
Wed Oct 01 00:00:00 EDT 2014
Chondroprotective effects and multi-target mechanisms of Icariin in IL-1 beta-induced human SW 1353 chondrosarcoma cells and a rat osteoarthritis model.
International immunopharmacology
Zeng L,Wang W,Rong XF,Zhong Y,Jia P,Zhou GQ,Li RH
Wed Jan 01 00:00:00 EST 2014
Chondroprotective effects and multi-target mechanisms of Icariin in IL-1 beta-induced human SW 1353 chondrosarcoma cells and a rat osteoarthritis model.
International immunopharmacology
Zeng L,Wang W,Rong XF,Zhong Y,Jia P,Zhou GQ,Li RH
Wed Jan 01 00:00:00 EST 2014
Gemcitabine resistance in breast cancer cells regulated by PI3K/AKT-mediated cellular proliferation exerts negative feedback via the MEK/MAPK and mTOR pathways.
OncoTargets and therapy
Yang XL,Lin FJ,Guo YJ,Shao ZM,Ou ZL
Thu Jun 26 00:00:00 EDT 2014
Gemcitabine resistance in breast cancer cells regulated by PI3K/AKT-mediated cellular proliferation exerts negative feedback via the MEK/MAPK and mTOR pathways.
OncoTargets and therapy
Yang XL,Lin FJ,Guo YJ,Shao ZM,Ou ZL
Thu Jun 26 00:00:00 EDT 2014
miR-203 inhibits tumor cell migration and invasion via caveolin-1 in pancreatic cancer cells.
Oncology letters
Miao L,Xiong X,Lin Y,Cheng Y,Lu J,Zhang J,Cheng N
Sat Mar 01 00:00:00 EST 2014
miR-203 inhibits tumor cell migration and invasion via caveolin-1 in pancreatic cancer cells.
Oncology letters
Miao L,Xiong X,Lin Y,Cheng Y,Lu J,Zhang J,Cheng N
Sat Mar 01 00:00:00 EST 2014
The crucial role of Atg5 in cortical neurogenesis during early brain development.
Scientific reports
Lv X,Jiang H,Li B,Liang Q,Wang S,Zhao Q,Jiao J
Mon Aug 11 00:00:00 EDT 2014
The crucial role of Atg5 in cortical neurogenesis during early brain development.
Scientific reports
Lv X,Jiang H,Li B,Liang Q,Wang S,Zhao Q,Jiao J
Mon Aug 11 00:00:00 EDT 2014
MiR-424-5p reversed epithelial-mesenchymal transition of anchorage-independent HCC cells by directly targeting ICAT and suppressed HCC progression.
Scientific reports
Zhang Y,Li T,Guo P,Kang J,Wei Q,Jia X,Zhao W,Huai W,Qiu Y,Sun L,Han L
Mon Sep 01 00:00:00 EDT 2014
MiR-424-5p reversed epithelial-mesenchymal transition of anchorage-independent HCC cells by directly targeting ICAT and suppressed HCC progression.
Scientific reports
Zhang Y,Li T,Guo P,Kang J,Wei Q,Jia X,Zhao W,Huai W,Qiu Y,Sun L,Han L
Mon Sep 01 00:00:00 EDT 2014
MiR-424-5p reversed epithelial-mesenchymal transition of anchorage-independent HCC cells by directly targeting ICAT and suppressed HCC progression.
Scientific reports
Zhang Y,Li T,Guo P,Kang J,Wei Q,Jia X,Zhao W,Huai W,Qiu Y,Sun L,Han L
Mon Sep 01 00:00:00 EDT 2014
MiR-424-5p reversed epithelial-mesenchymal transition of anchorage-independent HCC cells by directly targeting ICAT and suppressed HCC progression.
Scientific reports
Zhang Y,Li T,Guo P,Kang J,Wei Q,Jia X,Zhao W,Huai W,Qiu Y,Sun L,Han L
Mon Sep 01 00:00:00 EDT 2014
FNC, a novel nucleoside analogue, blocks invasion of aggressive non-Hodgkin lymphoma cell lines via inhibition of the Wnt/β-catenin signaling pathway.
Asian Pacific journal of cancer prevention : APJCP
Zhang Y,Wang CP,Ding XX,Wang N,Ma F,Jiang JH,Wang QD,Chang JB
Mon Jun 08 00:00:00 EDT 2015
FNC, a novel nucleoside analogue, blocks invasion of aggressive non-Hodgkin lymphoma cell lines via inhibition of the Wnt/β-catenin signaling pathway.
Asian Pacific journal of cancer prevention : APJCP
Zhang Y,Wang CP,Ding XX,Wang N,Ma F,Jiang JH,Wang QD,Chang JB
Mon Jun 08 00:00:00 EDT 2015
Role of Wnt/β-catenin signaling pathway in the mechanism of calcification of aortic valve.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Gu GJ,Chen T,Zhou HM,Sun KX,Li J
Sat Feb 01 00:00:00 EST 2014
Role of Wnt/β-catenin signaling pathway in the mechanism of calcification of aortic valve.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Gu GJ,Chen T,Zhou HM,Sun KX,Li J
Sat Feb 01 00:00:00 EST 2014
Overexpression of JAKMIP1 associates with Wnt/β-catenin pathway activation and promotes cancer cell proliferation in vitro.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Okai I,Wang L,Gong L,Arko-Boham B,Hao L,Zhou X,Qi X,Hu J,Shao S
Mon Apr 01 00:00:00 EDT 2013
Overexpression of JAKMIP1 associates with Wnt/β-catenin pathway activation and promotes cancer cell proliferation in vitro.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Okai I,Wang L,Gong L,Arko-Boham B,Hao L,Zhou X,Qi X,Hu J,Shao S
Mon Apr 01 00:00:00 EDT 2013
Caudatin inhibits human hepatoma cell growth and metastasis through modulation of the Wnt/β-catenin pathway.
Oncology reports
Luo Y,Sun Z,Li Y,Liu L,Cai X,Li Z
Sun Dec 01 00:00:00 EST 2013
Caudatin inhibits human hepatoma cell growth and metastasis through modulation of the Wnt/β-catenin pathway.
Oncology reports
Luo Y,Sun Z,Li Y,Liu L,Cai X,Li Z
Sun Dec 01 00:00:00 EST 2013
Vascular endothelial growth factor receptor-1 activation promotes migration and invasion of breast cancer cells through epithelial-mesenchymal transition.
PloS one
Ning Q,Liu C,Hou L,Meng M,Zhang X,Luo M,Shao S,Zuo X,Zhao X
Thu Jan 16 00:00:00 EST 2014
Vascular endothelial growth factor receptor-1 activation promotes migration and invasion of breast cancer cells through epithelial-mesenchymal transition.
PloS one
Ning Q,Liu C,Hou L,Meng M,Zhang X,Luo M,Shao S,Zuo X,Zhao X
Thu Jan 16 00:00:00 EST 2014
Caveolin-1 knockdown is associated with the metastasis and proliferation of human lung cancer cell line NCI-H460.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Song Y,Xue L,Du S,Sun M,Hu J,Hao L,Gong L,Yeh D,Xiong H,Shao S
Sat Sep 01 00:00:00 EDT 2012
Caveolin-1 knockdown is associated with the metastasis and proliferation of human lung cancer cell line NCI-H460.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Song Y,Xue L,Du S,Sun M,Hu J,Hao L,Gong L,Yeh D,Xiong H,Shao S
Sat Sep 01 00:00:00 EDT 2012
Influence of β-catenin small interfering RNA on human osteosarcoma cells.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Zhang F,Chen A,Chen J,Yu T,Guo F
Wed Jun 01 00:00:00 EDT 2011
Influence of β-catenin small interfering RNA on human osteosarcoma cells.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban
Zhang F,Chen A,Chen J,Yu T,Guo F
Wed Jun 01 00:00:00 EDT 2011
SiRNA-mediated silencing of beta-catenin suppresses invasion and chemosensitivity to doxorubicin in MG-63 osteosarcoma cells.
Asian Pacific journal of cancer prevention : APJCP
Zhang F,Chen A,Chen J,Yu T,Guo F
Wed May 30 00:00:00 EDT 2012
SiRNA-mediated silencing of beta-catenin suppresses invasion and chemosensitivity to doxorubicin in MG-63 osteosarcoma cells.
Asian Pacific journal of cancer prevention : APJCP
Zhang F,Chen A,Chen J,Yu T,Guo F
Wed May 30 00:00:00 EDT 2012