Stealth RNAi™ siRNA
Stealth RNAi™ siRNA is the next generation synthetic RNA from Invitrogen. The Stealth RNAi™ siRNA
molecules are 25 base-pair double-stranded RNA oligonucleotides with
proprietary chemical modifications.
Using
Stealth RNAi™ siRNA for your RNAi studies offers the following advantages over
traditional siRNA:
·
Decreased nonspecific
stress responses for clearer results
·
High potency for more
powerful knockdown of genes
·
More stable in both
cell culture systems and in vivo
For
additional product information, click Stealth RNAi™ siRNA
Stealth RNAi™ siRNA
The
BLOCK-iT™ RNAi Designer helps you find gene-specific targets and design 25
nucleotide Stealth RNAi™ siRNA molecules. When you input an accession number or a
nucleotide sequence, the designer:
·
Checks input sequence
for SNPs (if input is a RefSeq accession number)
·
BLAST input sequence
against databases and selects a number of unique regions
·
Uses proprietary design
rules* to find 25 nucleotide gene-specific targets for RNAi analysis and
reports up to 10 top scoring Stealth RNAi™ siRNA targets
See
the Online Help for additional information.
Stealth RNAi™ siRNA Ordering
You
can order Stealth RNAi™ siRNA via the RNAi Designer and 5nmoles of 25 nt Stealth RNAi™ siRNA molecules arrive duplexed and desalted. Check ‘Custom Primer
Resources’ for other ordering options.
Proprietary design
rules*: The proprietary design rules are derived based on statistical analysis
of RNA experimental data (e.g. sequence composition, nucleotide content at 5'
and 3' ends, thermodynamic properties) collected from multiple validated
(functional and non-functional) RNA oligo sets tested on different gene
targets.