A 4-nucleotide, 5' overhang (TGCT) complementary to the vector for directional cloning
A 5' G + the 21-nucleotide antisense sequence derived from the target gene, followed by
A short spacer of 19 nucleotides to form a terminal loop and
A short sense target sequence with 2 nucleotides removed to create an internal loop
When you are ready to order, select the checkbox next to each design that you want to order and click on Go to Order Form to proceed to ordering.