The BLOCK-iT™ RNAi Designer will generate up to 10 designs per target sequence. Each design is given one of the following five Probability rankings:
Designs with this ranking closely
fit the characteristics of the most effective RNAi molecules. They have
the greatest likelihood of resulting in >70% knockdown. We recommend
testing 2 or 3 designs from this group.
Designs with this ranking fit
most of the characteristics of effective RNAi molecules. They are very
likely to result in >70% effective knockdown. We recommend testing
3 designs from this group.
Designs with this ranking fit
some of the characteristics of effective RNAi molecules. They are likely
to result in >70% effective knockdown. We recommend testing at least
3 designs from this group.
Designs with this ranking have
fewer of the characteristics of effective RNAi molecules but they are
still likely to result in >70% effective knockdown. We recommend testing
at least 3 designs from this group.
Designs with this ranking have
the fewest characteristics of effective RNAi molecules but many are still
likely to result in >70% effective knockdown. We recommend testing
at least 3 designs from this group.