shRNA Final
Designs
This screen shows the final dsDNA oligo
designs. Each design includes a 4-nucleotide sequence (CACC or AAAA) for
directional cloning into the selected Gateway®-adapted entry vector, the
sense or antisense sequence beginning with the transcription initiation
site, the loop sequence you selected, and the complementary sense or antisense
sequence.
Top and bottom strands are listed separately
(5' to 3'), as is the double-stranded oligo required for directional cloning.
When you are ready to order, select the checkbox
next to each design that you want to order and click on Go
to Order Form to proceed to ordering.