siRNA Negative Control Sequences
An siRNA negative control sequence can be used in your experiments to detect nonspecific effects related to siRNA transfection. The Designer generates a separate negative control sequence for each siRNA oligo that you design. Each negative control design has no known homology to the target sequence that you provide. Note that only one negative control is required for each target sequence.
Each control sequence is listed in the table with its siRNA counterpart. Select the checkbox next to the control sequence that you want to order and click on Go to Order Form to proceed to ordering.