Stealth RNA™ siRNA Negative Control Sequences
A Stealth RNA™ siRNA negative control sequence can be used in your experiments to detect nonspecific effects related to transfection. The Designer generates a separate negative control sequence for each Stealth RNA™ siRNA oligo that you design. Each negative control design has no known homology to the target sequence that you provide. Note that only one negative control is required for each target sequence.
Note: A Universal Stealth RNA™ siRNA Negative Control is available separately from Invitrogen. Visit the Invitrogen online catalog for more information.
Each control sequence is listed in the table with its Stealth RNA™ siRNA counterpart. Select the checkbox next to the control sequence that you want to order and click on Go to Order Form to proceed to ordering.