BLOCK-iT™ RNAi Express
READY, CLICK and GO: Fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks.
 RNAi Express Options:  Search By ID/Keyword Browse By Pathways Browse By Gene Families
MAP Kinase Pathway
Mitogen-activated protein kinases (MAPK) are a family of serine/threonine kinases that respond to diverse stimuli within a tiered phosphorelay system (Figure 1). MAPK activation can occur in response to growth factors, differentiation agents, hormones, cytokines, and environmental stress. Pathway specificity is achieved through the integration of distinct signaling modules comprised of specific MAPK subfamilies including: extracellular signal regulated kinase (ERK1/2), c-Jun N-terminal kinase (JNK1/2/3), p38 proteins (p38α,β,γ,δ), and Big MAPK (BMK/ERK5). Each of these MAPK subtype containing modules consists of a three tier regulatory cascade of MAPKKKs, MAPKKs, and MAPKs. MAPKKKs phosphorylate MAPKKs on serine and threonine residues in response to cellular activation, often through direct interaction with small GTPases or kinases (MAPKKKKs). These dual specificity MAPKKs (also known as MEKs or MKKs) phosphorylate specific MAPKs on conserved Thr and Tyr residues. Following activation, MAPKs catalyze the phosphorylation of other protein kinases, phospholipases, transcription factors, and cytoskeletal proteins leading to changes in gene transcription, protein synthesis, metabolism, cell survival, cell proliferation, and cell motility.
MAP Kinase Pathway Validated Stealth RNAi™ siRNA
MAPK
MAPKK
MAPKKK
Upstream Regulators of MAPK
Scaffolding Proteins
Targets of MAPK
Figure 1. Organization of MAP Kinase Signal Transduction Cascade
References
1. Chang, L & Karin, M. Mammalian MAP kinase signalling cascades. Nature 410, 37-40 (2001).
2. Davis R.J. Signal Transduction by the JNK group of MAP kinases. Cell 103, 239-252 (2000).
3. Garrington, T.P. & Johnson, G.L. Organization and regulation of mitogen-activated protein kinase signaling pathways. Curr. Opin. Cell Biol. 11, 211-218 (1999).
4. Hazzalin, C.A. & Mahadevan, L.C. MAPK-regulated transcription: a continuously variable gene switch? Nat. Rev. Mol. Cell Biol. 3, 30-40 (2002).
5. Johnson, G.L. & Lapadat R. Mitogen-activated protein kinase pathways mediated by ERK, JNK, and p38 protein kinases. Science 298, 1911-1912 (2002).
6. Kumar et al. P38 MAP Kinases: Key signaling molecules as therapeutic targets for inflammatory diseases. Nat. Rev. Drug Discov. 2, 717-726 (2003).
7. Kyriakis, J.M. & Avruch, J. Mammalian mitogen-activated protein kinase signal transduction pathways activated by stress and inflammation. Physiol. Rev. 81, 807-869 (2001).
8. Manning, A.M. & Davis R.J. Target JNK for therapeutic benefit: from junk to gold? Nat. Rev. Drug Discov. 2, 554-65 (2003).
9. Morrison, D.K. & Davis, R.J. Regulation of MAP kinase signaling modules by scaffold proteins in mammals Ann. Rev. Cell Dev. Biol. 19, 91-118 (2003).
10. Yang, S.H., Sharrocks, A.D. & Whitmarsh, A.J. Transcription regulation by MAP kinase signaling cascades. Gene 320, 3-21 (2003).
test
×
Please sign in before using our BLOCK-iT RNAi Designer to ensure the best experience