Protocol
for Annealing for dsRNA
- Resuspend
each RNA oligo to a concentration of 50 µM.
- Combine 30 µl of each RNA oligo solution
and 15 µl of 5X Annealing Buffer. Final volume is 75 µl.
- Incubate the solution for 1 minute at 90°
C .
- Centrifuge the tube for 15 seconds to
bring the solution to the bottom.
- Allow to cool slowly to room temperature.
(This is best accomplished using a beaker of water as a waterbath to
achieve the 90°C temperature and then removing the heat source and
allowing the beaker of water containing the tube of oligos to cool slowly
to room temp.)
- The solution can be stored frozen at 20°
C and freeze-thawed up to 5 times. The final concentration of siRNA duplex
is 20 µMolar.
5X
Annealing Buffer:
50 mM Tris-HCl, pH 8.0
100 mM NaCl
5 mM EDTA
For
additional information, click RNA Oligo Supporting
Information.