siRNA
Invitrogen offers standard siRNA with the following options:
- single-stranded oligos or duplex siRNA
- deprotected and desalted
- standard sized (21-23 nt) or custom sized
(10-50 nt) with 3’ TT overhang
- 50 and 200 nmoles
- HPLC purification by request and quality
control by mass spectrometry
The
turn-around time for single-stranded desalted is 5-7 days and HPLC-purified is
7-9 days. The turn-around time for double-stranded desalted is 6-8 days and
HPLC-purified is 8-10 days.
For
additional product information, click BLOCK-iT™ siRNA
and FAQs.
siRNA Design
The
BLOCK-iT™ RNAi Designer helps you find gene-specific targets and design
21-nucleotide siRNA. When you input an accession number or a nucleotide
sequence, the designer:
·
Checks input sequence
for SNPs (if input is a RefSeq accession number)
·
BLAST input sequence
against databases and selects a number of unique regions
·
Uses proprietary design
rules* to find 21 nucleotide gene-specific targets for RNAi analysis and
reports up to 10 top scoring siRNA targets.
You have an option to select two different motif patterns for siRNA design:
Default motif pattern:
If you select ‘Default motif pattern’, the RNAi Designer finds all 19 nt target
sequences that fulfill proprietary design criteria and reports up to top 10
high scoring target sequences. You can order siRNA oligos for selected target
sequences via the RNAi Designer. The results using ‘Default motif pattern’ may
contain Tuschl’s patterns, but if you are specifically looking for Tuschl’s
patterns, select ‘Tushcl’s motif Pattern’ as your option.
Tuschl’s motif pattern:
If you select ‘Tuschl’s motif pattern’, the RNAi Designer finds 19 nt target
sequences that fulfill the selected pattern (nucleotide composition of 19 nt
and –1, -2 and 20, 21 base position composition on the target sequence) and
proprietary design criteria. The designer reports up to 10 top scoring target sequences
and you can order siRNA oligos for selected target sequences via the designer.
We recommend using ‘Default motif pattern’ over ‘Tuschl’s motif pattern’, as
the stringency of selection is much higher when ‘Default motif pattern’ is
selected.
References:
·
Elbashir SM et al. (2001) Duplexes of
21-nucleotide RNAs mediate RNA interference in cultured mammalian cells.
Nature. 411:494-498.
·
Elbahir SM et al. (2001). Functional anatomy of
siRNAs for mediating efficient RNAi in Drosophila melanogaster embryo lysate.
EMBO J. 20:6877-6888.
·
Elbashir SM et al. (2002). Analysis of gene
function in somatic mammalian cells using small interfering RNAs. Methods.
26:199-213
See
Online Help for additional information.
siRNA Ordering
You
can order siRNA via the RNAi Designer and check ‘Custom Primer
Resources’ for other ordering options.
Proprietary design rules*:
The proprietary design rules are derived based on statistical analysis of RNA
experimental data (e.g. sequence composition, nucleotide content at 5' and 3'
ends, thermodynamic properties) collected from multiple validated (functional
and non-functional) RNA oligo sets tested on different gene targets.