miR RNAi Design Results
In this screen, you select from a list of top-scoring sequences derived from the target sequence that will be used to design your miR RNAi oligos. The sequences have been selected based on a statistical analysis of multiple validated miR RNAi sequences and a proprietary algorithm.
Sequence Information
Information about the target sequence is listed at the top of the page. If you copied and pasted a nucleotide sequence, only the sequence length will be listed. Information about the gene is included if you entered an accession number.
Results Table
To view the results of the BLAST search selected in Step 3 of the previous page, including which regions of the sequence were masked due to known homologies, click on View BLAST Results.
In the results table, each design is listed with a selection checkbox. If you entered an accession number for the target sequence, the Region of the gene for each design will be listed. The GC% column lists the GC percentage of the design. Finally, the design ranking (in stars) is listed next to each design.
Note: We recommend selecting 2–4 designs for experimental testing; the number of designs you select will depend on their rankings.
Select the checkbox next to each design that you want to order and click on Design miR RNAi. In the following screen, the dsDNA oligo sequence will be displayed for final selection.
If the Designer is Unable to Generate Suitable Designs
If the Designer is unable to generate a suitable number of miR RNAi designs for your target sequence, you can click on the Back to RNAi Design Page link to return to the design parameters screen. You can then change some or all of the following parameters: