Select the shRNA Loop Sequence
In this screen, you select the nucleotide spacer between the sense and antisense sequences that will generate the loop structure of each shRNA oligo.
Select one of the predesigned 4-nucleotide loop sequences from the Default Loop Sequence pulldown list, or enter a custom loop sequence of 4–11 nucleotides in the Custom Loop Sequence field. Note that short loop sequences (4–7 nucleotides) are usually more efficient. Avoid using a loop sequence containing thymidines (T’s) as they may cause early termination. This is particularly true if the target sequence ends in a T residue.
When you have specified a loop sequence for each target, click on the Design button to generate the final shRNA designs. See shRNA Final Designs for more information.