Stealth RNA™ siRNA Design Results
In this screen, you select from a list of up to 10 top-scoring Stealth RNA™ siRNA designs generated from your target sequence. The designs have been generated from a statistical analysis of multiple validated Stealth RNA™ siRNA sets and a proprietary algorithm.
Sequence Information
Information about the target sequence is listed at the top of the page. If you copied and pasted a nucleotide sequence, only the sequence length will be listed. Information about the gene is included if you entered an accession number.
Results Table
To view the results of the BLAST search selected in Step 3 of the previous page, including which regions of the sequence were masked due to known homologies, click on View BLAST Results.
In the results table, each design is listed with a selection checkbox. If you entered an accession number for the target sequence, the Gene Region of each design will be listed. The GC% column lists the GC percentage of the design. The Tuschl's Pattern Match column lists the Tuschl's patterns that the design matches. Finally, the probability of experimental success is listed next to each design.
Note: We recommend selecting 2–4 designs for experimental testing; the number of designs you select will depend on their probability rankings.
Select the checkbox next to each design that you want to order and click on Continue. In the following screen, you will be prompted to select any Negative Control Sequences that you want to order with your Stealth RNA™ siRNA molecules.
If the Designer is Unable to Generate Suitable Designs
If the Designer is unable to generate a suitable number of designs for your target sequence, you can click on the Back button on your browser to return to the design parameters screen. You can then change some or all of the following design parameters: