Product References
Toxin expression during Staphylococcus aureus infection imprints host immunity to inhibit vaccine efficacy.
NPJ vaccines
Teymournejad O,Li Z,Beesetty P,Yang C,Montgomery CP
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Jan 24 00:00:00 EST 2023
NCR negative group 3 innate lymphoid cell (NCR- ILC3) participates in abnormal pathology of lung in cigarette smoking-induced COPD mice.
Immunity, inflammation and disease
Chu S,Ma L,Yang X,Xiao B,Liang Y,Zheng S,Li L
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Adipocyte-Secreted IL-6 Sensitizes Macrophages to IL-4 Signaling.
Diabetes
Luan D,Dadpey B,Zaid J,Bridge-Comer PE,DeLuca JH,Xia W,Castle J,Reilly SM
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
A novel ZsGreen knock-in melanoma cell line reveals the function of CD11b in tumor phagocytosis.
Immunology and cell biology
Zhang L,Feng X,Shen Y,Wang Y,Liu Z,Ma Y,Gu Y,Guo G,Duan L,Lu L,Liang Y,Lawrence T,Huang R
35-0031-82 was used in Flow cytometry/Cell sorting to demonstrate that the ZsGreen-expressing YUMM1.7 melanoma model provides a valuable tool for the study of phagocytosis in vivo.
Sat Oct 01 00:00:00 EDT 2022
Loss of Optineurin Drives Cancer Immune Evasion via Palmitoylation-Dependent IFNGR1 Lysosomal Sorting and Degradation.
Cancer discovery
Du W,Hua F,Li X,Zhang J,Li S,Wang W,Zhou J,Wang W,Liao P,Yan Y,Li G,Wei S,Grove S,Vatan L,Zgodziński W,Majewski M,Wallner G,Chen H,Kryczek I,Fang JY,Zou W
35-0031-82 was used in Flow Cytometry to show that optineurin is a shared node between the IFN and MHC pathways and predicts colorectal cancer patient outcome.
Thu Jul 01 00:00:00 EDT 2021
Fasudil prevents liver fibrosis via activating natural killer cells and suppressing hepatic stellate cells.
World journal of gastroenterology
Han QJ,Mu YL,Zhao HJ,Zhao RR,Guo QJ,Su YH,Zhang J
35-0031-82 was used in Flow cytometry/Cell sorting to evaluate the anti-fibrotic effects of Fasudil in a mouse model of liver fibrosis induced by thioacetamide.
Mon Jun 28 00:00:00 EDT 2021
A novel Cytochrome P450 26A1 expressing NK cell subset at the mouse maternal-foetal interface.
Journal of cellular and molecular medicine
Wei DP,Li DD,Gu AQ,Ji WH,Yang Y,Peng JP
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Construction of a 3D brain extracellular matrix model to study the interaction between microglia and T cells in co-culture.
The European journal of neuroscience
Frühauf M,Zeitschel U,Höfling C,Ullm F,Rabiger FV,Alber G,Pompe T,Müller U,Roßner S
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Imbalance between T helper 1 and regulatory T cells plays a detrimental role in experimental Parkinson's disease in mice.
The Journal of international medical research
Li W,Luo Y,Xu H,Ma Q,Yao Q
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Apr 01 00:00:00 EDT 2021
Effective delivery of STING agonist using exosomes suppresses tumor growth and enhances antitumor immunity.
The Journal of biological chemistry
McAndrews KM,Che SPY,LeBleu VS,Kalluri R
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Aug 20 00:00:00 EDT 2021
The effect of fingolimod on regulatory T cells in a mouse model of brain ischaemia.
Journal of neuroinflammation
Malone K,Diaz Diaz AC,Shearer JA,Moore AC,Waeber C
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Sat Jan 30 00:00:00 EST 2021
Obesity results in adipose tissue T cell exhaustion.
JCI insight
Porsche CE,Delproposto JB,Geletka L,O'Rourke R,Lumeng CN
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Apr 22 00:00:00 EDT 2021
Theranostic near-infrared-IIb emitting nanoprobes for promoting immunogenic radiotherapy and abscopal effects against cancer metastasis.
Nature communications
Li H,Wang M,Huang B,Zhu SW,Zhou JJ,Chen DR,Cui R,Zhang M,Sun ZJ
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Dec 09 00:00:00 EST 2021
Hypoxia-sensing CAR T cells provide safety and efficacy in treating solid tumors.
Cell reports. Medicine
Kosti P,Opzoomer JW,Larios-Martinez KI,Henley-Smith R,Scudamore CL,Okesola M,Taher MYM,Davies DM,Muliaditan T,Larcombe-Young D,Woodman N,Gillett CE,Thavaraj S,Maher J,Arnold JN
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Apr 20 00:00:00 EDT 2021
Loss of fragile site-associated tumor suppressor promotes antitumor immunity via macrophage polarization.
Nature communications
Zhang L,Zhang K,Zhang J,Zhu J,Xi Q,Wang H,Zhang Z,Cheng Y,Yang G,Liu H,Guo X,Zhou D,Xue Z,Li Y,Zhang Q,Da Y,Liu L,Yin Z,Yao Z,Zhang R
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Jul 14 00:00:00 EDT 2021
Histological, behavioural and flow cytometric datasets relating to acute ischaemic stroke in young, aged and ApoE-/- mice in the presence and absence of immunomodulation with fingolimod.
Data in brief
Diaz Diaz AC,Malone K,Shearer JA,Moore AC,Waeber C
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Targeting Ovarian Carcinoma with TSP-1:CD47 Antagonist TAX2 Activates Anti-Tumor Immunity.
Cancers
Jeanne A,Sarazin T,Charlé M,Moali C,Fichel C,Boulagnon-Rombi C,Callewaert M,Andry MC,Diesis E,Delolme F,Rioult D,Dedieu S
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Oct 07 00:00:00 EDT 2021
A nanounit strategy reverses immune suppression of exosomal PD-L1 and is associated with enhanced ferroptosis.
Nature communications
Wang G,Xie L,Li B,Sang W,Yan J,Li J,Tian H,Li W,Zhang Z,Tian Y,Dai Y
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Sep 30 00:00:00 EDT 2021
Non-redundant activity of GSK-3α and GSK-3β in T cell-mediated tumor rejection.
iScience
Steele L,Mannion AJ,Shaw G,Maclennan KA,Cook GP,Rudd CE,Taylor A
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Jun 25 00:00:00 EDT 2021
Excessive Costimulation Leads to Dysfunction of Adoptively Transferred T Cells.
Cancer immunology research
Wijewarnasuriya D,Bebernitz C,Lopez AV,Rafiq S,Brentjens RJ
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
Antitumor Effect and Immune Response of Nanosecond Pulsed Electric Fields in Pancreatic Cancer.
Frontiers in oncology
Zhao J,Chen S,Zhu L,Zhang L,Liu J,Xu D,Tian G,Jiang T
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Oct 28 00:00:00 EDT 2022
Inhibition of Glycolysis in Pathogenic TH17 Cells through Targeting a miR -21-Peli1-c-Rel Pathway Prevents Autoimmunity.
Journal of immunology (Baltimore, Md. : 1950)
Qiu R,Yu X,Wang L,Han Z,Yao C,Cui Y,Hou G,Dai D,Jin W,Shen N
35-0031-82 was used in Flow Cytometry to investigate if CNS-infiltrated pathogenic TH17 cells from diseased mice specifically upregulate glycolytic pathway genes compared with homeostatic intestinal TH17 cells.
Mon Jun 15 00:00:00 EDT 2020
Enhanced alleviation of aGVHD by TGF-β1-modified mesenchymal stem cells in mice through shifting MΦ into M2 phenotype and promoting the differentiation of Treg cells.
Journal of cellular and molecular medicine
Wu R,Liu C,Deng X,Chen L,Hao S,Ma L
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
DEK-targeting aptamer DTA-64 attenuates bronchial EMT-mediated airway remodelling by suppressing TGF-β1/Smad, MAPK and PI3K signalling pathway in asthma.
Journal of cellular and molecular medicine
Song Y,Wang Z,Jiang J,Piao Y,Li L,Xu C,Piao H,Li L,Yan G
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
Impact of S1P Mimetics on Mesenteric Ischemia/Reperfusion Injury.
Pharmaceuticals (Basel, Switzerland)
Potì F,Giorgio C,Zini I,Nofer JR,Vivo V,Palese S,Ballabeni V,Barocelli E,Bertoni S
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Oct 09 00:00:00 EDT 2020
CD279 mediates the homeostasis and survival of regulatory T cells by enhancing T cell and macrophage interactions.
FEBS open bio
Liu Y,Gao Y,Hao H,Hou T
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2020
TNFAIP3 Plays a Role in Aging of the Hematopoietic System.
Frontiers in immunology
Smith MA,Culver-Cochran AE,Adelman ER,Rhyasen GW,Ma A,Figueroa ME,Starczynowski DT
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Apr 28 00:00:00 EDT 2021
IDO1+ Paneth cells promote immune escape of colorectal cancer.
Communications biology
Pflügler S,Svinka J,Scharf I,Crncec I,Filipits M,Charoentong P,Tschurtschenthaler M,Kenner L,Awad M,Stift J,Schernthanner M,Bischl R,Herndler-Brandstetter D,Glitzner E,Moll HP,Casanova E,Timelthaler G,Sibilia M,Gnant M,Lax S,Thaler J,Müller M,Strobl B,Mohr T,Kaser A,Trajanoski Z,Heller G,Eferl R
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri May 22 00:00:00 EDT 2020
IDO1+ Paneth cells promote immune escape of colorectal cancer.
Communications biology
Pflügler S,Svinka J,Scharf I,Crncec I,Filipits M,Charoentong P,Tschurtschenthaler M,Kenner L,Awad M,Stift J,Schernthanner M,Bischl R,Herndler-Brandstetter D,Glitzner E,Moll HP,Casanova E,Timelthaler G,Sibilia M,Gnant M,Lax S,Thaler J,Müller M,Strobl B,Mohr T,Kaser A,Trajanoski Z,Heller G,Eferl R
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri May 22 00:00:00 EDT 2020
TET2-Loss-of-Function-Driven Clonal Hematopoiesis Exacerbates Experimental Insulin Resistance in Aging and Obesity.
Cell reports
Fuster JJ,Zuriaga MA,Zorita V,MacLauchlan S,Polackal MN,Viana-Huete V,Ferrer-Pérez A,Matesanz N,Herrero-Cervera A,Sano S,Cooper MA,González-Navarro H,Walsh K
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Oct 27 00:00:00 EDT 2020
Artificial Mini Dendritic Cells Boost T Cell-Based Immunotherapy for Ovarian Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Cheng S,Xu C,Jin Y,Li Y,Zhong C,Ma J,Yang J,Zhang N,Li Y,Wang C,Yang Z,Wang Y
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
CNS Immune Profiling in a Dengue Virus-Infected Immunocompetent Outbred ICR Mice Strain.
Frontiers in cellular and infection microbiology
Shen TJ,Chen CL,Jhan MK,Tseng PC,Lin CF
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Jun 17 00:00:00 EDT 2021
Tumor microenvironment remodeling by an engineered oncolytic adenovirus results in improved outcome from PD-L1 inhibition.
Oncoimmunology
Cervera-Carrascon V,Quixabeira DCA,Santos JM,Havunen R,Zafar S,Hemminki O,Heiniö C,Munaro E,Siurala M,Sorsa S,Mirtti T,Järvinen P,Mildh M,Nisen H,Rannikko A,Anttila M,Kanerva A,Hemminki A
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri May 22 00:00:00 EDT 2020
Critical role of WNK1 in MYC-dependent early mouse thymocyte development.
eLife
Köchl R,Vanes L,Llorian Sopena M,Chakravarty P,Hartweger H,Fountain K,White A,Cowan J,Anderson G,Tybulewicz VL
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Oct 14 00:00:00 EDT 2020
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
The Innate Immune Response to Infection Induces Erythropoietin-Dependent Replenishment of the Dendritic Cell Compartment.
Frontiers in immunology
Einwächter H,Heiseke A,Schlitzer A,Gasteiger G,Adler H,Voehringer D,Manz MG,Ruzsics Z,Dölken L,Koszinowski UH,Sparwasser T,Reindl W,Jordan S
35-0031-82 was used in Flow Cytometry to investigate the molecular and cellular mechanisms of inflammation-induced dendritic cell generation.
Mon Apr 19 00:00:00 EDT 2021
Cooperation between Constitutive and Inducible Chemokines Enables T Cell Engraftment and Immune Attack in Solid Tumors.
Cancer cell
Dangaj D,Bruand M,Grimm AJ,Ronet C,Barras D,Duttagupta PA,Lanitis E,Duraiswamy J,Tanyi JL,Benencia F,Conejo-Garcia J,Ramay HR,Montone KT,Powell DJ,Gimotty PA,Facciabene A,Jackson DG,Weber JS,Rodig SJ,Hodi SF,Kandalaft LE,Irving M,Zhang L,Foukas P,Rusakiewicz S,Delorenzi M,Coukos G
35-0031-82 was used in Flow Cytometry to investigate the role of chemokines in regulating T cell accumulation in solid tumors.
Mon Jun 10 00:00:00 EDT 2019
1,2,3,4,6-Penta-O-galloyl-β-d-glucose modulates perivascular inflammation and prevents vascular dysfunction in angiotensin II-induced hypertension.
British journal of pharmacology
Mikolajczyk TP,Nosalski R,Skiba DS,Koziol J,Mazur M,Justo-Junior AS,Kowalczyk P,Kusmierczyk Z,Schramm-Luc A,Luc K,Maffia P,Graham D,Kiss AK,Naruszewicz M,Guzik TJ
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Sat Jun 01 00:00:00 EDT 2019
CD160 serves as a negative regulator of NKT cells in acute hepatic injury.
Nature communications
Kim TJ,Park G,Kim J,Lim SA,Kim J,Im K,Shin MH,Fu YX,Del Rio ML,Rodriguez-Barbosa JI,Yee C,Suh KS,Kim SJ,Ha SJ,Lee KM
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Jul 22 00:00:00 EDT 2019
Sex Differences in Mouse Popliteal Lymph Nodes.
Scientific reports
Dill-Garlow R,Chen KE,Walker AM
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Jan 30 00:00:00 EST 2019
Tpl2 Protects Against Fulminant Hepatitis Through Mobilization of Myeloid-Derived Suppressor Cells.
Frontiers in immunology
Xu J,Pei S,Wang Y,Liu J,Qian Y,Huang M,Zhang Y,Xiao Y
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Oct 14 00:00:00 EDT 2020
STAT1 signaling shields T cells from NK cell-mediated cytotoxicity.
Nature communications
Kang YH,Biswas A,Field M,Snapper SB
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Feb 22 00:00:00 EST 2019
Effector CD4 T cells with progenitor potential mediate chronic intestinal inflammation.
The Journal of experimental medicine
Shin B,Kress RL,Kramer PA,Darley-Usmar VM,Bellis SL,Harrington LE
35-0031 was used in Flow cytometry/Cell sorting to indicate that the chronicity of T cell-mediated inflammation is perpetuated by specific effector CD4 T cells with stem-like properties.
Mon Jul 02 00:00:00 EDT 2018
11 Beta-hydroxysteroid dehydrogenase type 1 regulates synovitis, joint destruction, and systemic bone loss in chronic polyarthritis.
Journal of autoimmunity
Hardy RS,Fenton C,Croft AP,Naylor AJ,Begum R,Desanti G,Buckley CD,Lavery G,Cooper MS,Raza K
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
Gli3 in fetal thymic epithelial cells promotes thymocyte positive selection and differentiation by repression of Shh.
Development (Cambridge, England)
Solanki A,Yanez DC,Ross S,Lau CI,Papaioannou E,Li J,Saldaña JI,Crompton T
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Jan 31 00:00:00 EST 2018
Granulocyte-macrophage colony-stimulating factor neuroprotective activities in Alzheimer's disease mice.
Journal of neuroimmunology
Kiyota T,Machhi J,Lu Y,Dyavarshetty B,Nemati M,Yokoyama I,Mosley RL,Gendelman HE
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Jun 15 00:00:00 EDT 2018
Sipa1 deficiency unleashes a host-immune mechanism eradicating chronic myelogenous leukemia-initiating cells.
Nature communications
Xu Y,Ikeda S,Sumida K,Yamamoto R,Tanaka H,Minato N
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Mar 02 00:00:00 EST 2018
A stochastic epigenetic switch controls the dynamics of T-cell lineage commitment.
eLife
Ng KK,Yui MA,Mehta A,Siu S,Irwin B,Pease S,Hirose S,Elowitz MB,Rothenberg EV,Kueh HY
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Nov 20 00:00:00 EST 2018
ADAMTS1 protease is required for a balanced immune cell repertoire and tumour inflammatory response.
Scientific reports
Rodríguez-Baena FJ,Redondo-García S,Peris-Torres C,Martino-Echarri E,Fernández-Rodríguez R,Plaza-Calonge MDC,Anderson P,Rodríguez-Manzaneque JC
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Aug 30 00:00:00 EDT 2018
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Dec 11 00:00:00 EST 2018
Anti-TNF and thiopurine therapy in pregnant IBD patients does not significantly alter a panel of B-cell and T-cell subsets in 1-year-old infants.
Clinical and translational gastroenterology
Kattah MG,Milush JM,Burt T,McCabe RP,Whang MI,Ma A,Mahadevan U
35-0031 was used in Flow cytometry/Cell sorting to hypothesize that the increased risk of infection is due to abnormal development of the newborn immune system.
Tue Apr 03 00:00:00 EDT 2018
Limited Foxp3+ Regulatory T Cells Response During Acute Trypanosoma cruzi Infection Is Required to Allow the Emergence of Robust Parasite-Specific CD8+ T Cell Immunity.
Frontiers in immunology
Araujo Furlan CL,Tosello Boari J,Rodriguez C,Canale FP,Fiocca Vernengo F,Boccardo S,Beccaria CG,Adoue V,Joffre O,Gruppi A,Montes CL,Acosta Rodriguez EV
35-0031 was used in Flow cytometry/Cell sorting to study the role of regulatory T cells during Trypanosoma cruzi infection.
Thu Sep 26 00:00:00 EDT 2019
Myc Cooperates with Ras by Programming Inflammation and Immune Suppression.
Cell
Kortlever RM,Sodir NM,Wilson CH,Burkhart DL,Pellegrinet L,Brown Swigart L,Littlewood TD,Evan GI
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Nov 30 00:00:00 EST 2017
Autoreactive T Cells and Chronic Fungal Infection Drive Esophageal Carcinogenesis.
Cell host & microbe
Zhu F,Willette-Brown J,Song NY,Lomada D,Song Y,Xue L,Gray Z,Zhao Z,Davis SR,Sun Z,Zhang P,Wu X,Zhan Q,Richie ER,Hu Y
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Apr 12 00:00:00 EDT 2017
Ubiquitination of hnRNPA1 by TRAF6 links chronic innate immune signaling with myelodysplasia.
Nature immunology
Fang J,Bolanos LC,Choi K,Liu X,Christie S,Akunuru S,Kumar R,Wang D,Chen X,Greis KD,Stoilov P,Filippi MD,Maciejewski JP,Garcia-Manero G,Weirauch MT,Salomonis N,Geiger H,Zheng Y,Starczynowski DT
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
A CCR4 antagonist reverses the tumor-promoting microenvironment of renal cancer.
The Journal of clinical investigation
Berlato C,Khan MN,Schioppa T,Thompson R,Maniati E,Montfort A,Jangani M,Canosa M,Kulbe H,Hagemann UB,Duncan AR,Fletcher L,Wilkinson RW,Powles T,Quezada SA,Balkwill FR
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
PTEN drives Th17 cell differentiation by preventing IL-2 production.
The Journal of experimental medicine
Kim HS,Jang SW,Lee W,Kim K,Sohn H,Hwang SS,Lee GR
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Nov 06 00:00:00 EST 2017
RIG-I antiviral signaling drives interleukin-23 production and psoriasis-like skin disease.
EMBO molecular medicine
Zhu H,Lou F,Yin Q,Gao Y,Sun Y,Bai J,Xu Z,Liu Z,Cai W,Ke F,Zhang L,Zhou H,Wang H,Wang G,Chen X,Zhang H,Wang Z,Ginhoux F,Lu C,Su B,Wang H
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
Betulin from Hedyotis hedyotidea ameliorates concanavalin A-induced and T cell-mediated autoimmune hepatitis in mice.
Acta pharmacologica Sinica
Zhou YQ,Weng XF,Dou R,Tan XS,Zhang TT,Fang JB,Wu XW
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
Cross-Presentation of Soluble and Cell-Associated Antigen by Murine Hepatocytes Is Enhanced by Collectrin Expression.
Journal of immunology (Baltimore, Md. : 1950)
Dolina JS,Cechova S,Rudy CK,Sung SJ,Tang WW,Lee J,Hahn YS,Le TH
35-0031 was used in Flow cytometry/Cell sorting to identify the functional role of collectrin in the liver.
Wed Mar 15 00:00:00 EDT 2017
Chemokine Receptor-Dependent Control of Skin Tissue-Resident Memory T Cell Formation.
Journal of immunology (Baltimore, Md. : 1950)
Zaid A,Hor JL,Christo SN,Groom JR,Heath WR,Mackay LK,Mueller SN
35-0031 was used in Flow cytometry/Cell sorting to examine the signals required for the migration of tissue-resident memory T cells within the constrained epidermal environment.
Sun Oct 01 00:00:00 EDT 2017
Cytochrome P450 26A1 modulates natural killer cells in mouse early pregnancy.
Journal of cellular and molecular medicine
Meng CY,Li ZY,Fang WN,Song ZH,Yang DD,Li DD,Yang Y,Peng JP
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Isolation of cell type-specific apoptotic bodies by fluorescence-activated cell sorting.
Scientific reports
Atkin-Smith GK,Paone S,Zanker DJ,Duan M,Phan TK,Chen W,Hulett MD,Poon IK
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Jan 06 00:00:00 EST 2017
The major targets of acute norovirus infection are immune cells in the gut-associated lymphoid tissue.
Nature microbiology
Grau KR,Roth AN,Zhu S,Hernandez A,Colliou N,DiVita BB,Philip DT,Riffe C,Giasson B,Wallet SM,Mohamadzadeh M,Karst SM
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
Intratumoral modulation of the inducible co-stimulator ICOS by recombinant oncolytic virus promotes systemic anti-tumour immunity.
Nature communications
Zamarin D,Holmgaard RB,Ricca J,Plitt T,Palese P,Sharma P,Merghoub T,Wolchok JD,Allison JP
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2017
ROCK regulates the intermittent mode of interstitial T cell migration in inflamed lungs.
Nature communications
Mrass P,Oruganti SR,Fricke GM,Tafoya J,Byrum JR,Yang L,Hamilton SL,Miller MJ,Moses ME,Cannon JL
35-0031 was used in Flow cytometry/Cell sorting to examine T cell migration in a mouse model of acute lung injury with two-photon imaging of intact lung tissue.
Wed Oct 18 00:00:00 EDT 2017
The efficacy of oral and subcutaneous antigen-specific immunotherapy in murine cow's milk- and peanut allergy models.
Clinical and translational allergy
Vonk MM,Wagenaar L,Pieters RHH,Knippels LMJ,Willemsen LEM,Smit JJ,van Esch BCAM,Garssen J
35-0031 was used in Flow cytometry/Cell sorting to compare oral immunotherapy and subcutaneous immunotherapy models for cows milk and peanut allergy.
Fri Apr 08 00:00:00 EDT 2022
Host-Protozoan Interactions Protect from Mucosal Infections through Activation of the Inflammasome.
Cell
Chudnovskiy A,Mortha A,Kana V,Kennard A,Ramirez JD,Rahman A,Remark R,Mogno I,Ng R,Gnjatic S,Amir ED,Solovyov A,Greenbaum B,Clemente J,Faith J,Belkaid Y,Grigg ME,Merad M
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Oct 06 00:00:00 EDT 2016
Lymphatic vessels regulate immune microenvironments in human and murine melanoma.
The Journal of clinical investigation
Lund AW,Wagner M,Fankhauser M,Steinskog ES,Broggi MA,Spranger S,Gajewski TF,Alitalo K,Eikesdal HP,Wiig H,Swartz MA
35-0031 was used in Flow cytometry/Cell sorting to suggest that the establishment of tumour-associated inflammation and immunity critically depends on lymphatic vessel remodelling and drainage.
Thu Sep 01 00:00:00 EDT 2016
CD8+ T cells drive autoimmune hematopoietic stem cell dysfunction and bone marrow failure.
Journal of autoimmunity
Gravano DM,Al-Kuhlani M,Davini D,Sanders PD,Manilay JO,Hoyer KK
35-0031 was used in Flow cytometry/Cell sorting to investigate whether CD8+ T cells play a non-redundant role in driving bone marrow failure.
Thu Dec 01 00:00:00 EST 2016
Autophagy deficiency in myeloid cells increases susceptibility to obesity-induced diabetes and experimental colitis.
Autophagy
Lee HY,Kim J,Quan W,Lee JC,Kim MS,Kim SH,Bae JW,Hur KY,Lee MS
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Aug 02 00:00:00 EDT 2016
Foxp3(+) T cells expressing RORγt represent a stable regulatory T-cell effector lineage with enhanced suppressive capacity during intestinal inflammation.
Mucosal immunology
Yang BH,Hagemann S,Mamareli P,Lauer U,Hoffmann U,Beckstette M,Föhse L,Prinz I,Pezoldt J,Suerbaum S,Sparwasser T,Hamann A,Floess S,Huehn J,Lochner M
35-0031 was used in Flow cytometry/Cell sorting to demonstrate that Foxp3(+) T cells expressing retinoic acid-related orphan receptor-γt, represent a stable regulatory T cell lineage with a highly suppressive phenotype in vivo.
Tue Mar 01 00:00:00 EST 2016
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Aug 15 00:00:00 EDT 2016
Protein Translation Activity: A New Measure of Host Immune Cell Activation.
Journal of immunology (Baltimore, Md. : 1950)
Seedhom MO,Hickman HD,Wei J,David A,Yewdell JW
35-0031 was used in Flow cytometry/Cell sorting to describe the in vivo ribopuromycylation method.
Mon Aug 15 00:00:00 EDT 2016
Treatment with Interleukin-7 Restores Host Defense against Pneumocystis in CD4+ T-Lymphocyte-Depleted Mice.
Infection and immunity
Ruan S,Samuelson DR,Assouline B,Morre M,Shellito JE
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Stromal senescence establishes an immunosuppressive microenvironment that drives tumorigenesis.
Nature communications
Ruhland MK,Loza AJ,Capietto AH,Luo X,Knolhoff BL,Flanagan KC,Belt BA,Alspach E,Leahy K,Luo J,Schaffer A,Edwards JR,Longmore G,Faccio R,DeNardo DG,Stewart SA
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Jun 08 00:00:00 EDT 2016
The dual role of NK cells in antitumor reactions triggered by ionizing radiation in combination with hyperthermia.
Oncoimmunology
Finkel P,Frey B,Mayer F,Bösl K,Werthmöller N,Mackensen A,Gaipl US,Ullrich E
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
The ER membrane-anchored ubiquitin ligase Hrd1 is a positive regulator of T-cell immunity.
Nature communications
Xu Y,Zhao F,Qiu Q,Chen K,Wei J,Kong Q,Gao B,Melo-Cardenas J,Zhang B,Zhang J,Song J,Zhang DD,Zhang J,Fan Y,Li H,Fang D
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Jul 15 00:00:00 EDT 2016
Irradiation of necrotic cancer cells, employed for pulsing dendritic cells (DCs), potentiates DC vaccine-induced antitumor immunity against high-grade glioma.
Oncoimmunology
Vandenberk L,Garg AD,Verschuere T,Koks C,Belmans J,Beullens M,Agostinis P,De Vleeschouwer S,Van Gool SW
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Photothermal therapy with immune-adjuvant nanoparticles together with checkpoint blockade for effective cancer immunotherapy.
Nature communications
Chen Q,Xu L,Liang C,Wang C,Peng R,Liu Z
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Oct 21 00:00:00 EDT 2016
Group 2 innate lymphoid cells promote beiging of white adipose tissue and limit obesity.
Nature
Brestoff JR,Kim BS,Saenz SA,Stine RR,Monticelli LA,Sonnenberg GF,Thome JJ,Farber DL,Lutfy K,Seale P,Artis D
35-0031 was used in Flow cytometry/Cell sorting to study how ILC2s can regulate adipose function and metabolic homeostasis.
Thu Mar 12 00:00:00 EDT 2015
Peroxisomal lipid synthesis regulates inflammation by sustaining neutrophil membrane phospholipid composition and viability.
Cell metabolism
Lodhi IJ,Wei X,Yin L,Feng C,Adak S,Abou-Ezzi G,Hsu FF,Link DC,Semenkovich CF
35-0031 was used in Flow cytometry/Cell sorting to investigate the immunometabolism pathway by which peroxisomal lipid synthesis regulates neutrophil development and inflammation.
Tue Jan 06 00:00:00 EST 2015
Nuclear export inhibitors avert progression in preclinical models of inflammatory demyelination.
Nature neuroscience
Haines JD,Herbin O,de la Hera B,Vidaurre OG,Moy GA,Sun Q,Fung HY,Albrecht S,Alexandropoulos K,McCauley D,Chook YM,Kuhlmann T,Kidd GJ,Shacham S,Casaccia P
35-0031 was used in Flow cytometry/Cell sorting to reveal that CRM1 inhibitors have the ability to modulate the export of molecules associated with both axonal damage and also neuroprotection.
Wed Apr 01 00:00:00 EDT 2015
α-Galactosylceramide suppresses murine eosinophil production through interferon-γ-dependent induction of NO synthase and CD95.
British journal of pharmacology
Gaspar-Elsas MI,Queto T,Masid-de-Brito D,Vieira BM,de Luca B,Cunha FQ,Xavier-Elsas P
35-0031 was used in Flow cytometry/Cell sorting to explore the haematopoietic effects of α-Galactosylceramide.
Wed Jul 01 00:00:00 EDT 2015
Novel mode of ISG15-mediated protection against influenza A virus and Sendai virus in mice.
Journal of virology
Morales DJ,Monte K,Sun L,Struckhoff JJ,Agapov E,Holtzman MJ,Stappenbeck TS,Lenschow DJ
35-0031 was used in Flow cytometry/Cell sorting to characterise the pathogenesis of influenza A virus and Sendai virus in ISG15(-/-) mice, showing that these mice are more susceptible to infection, and that ISG15 does not seem to directly inhibit virus replication or modulation of the acute immune response.
Thu Jan 01 00:00:00 EST 2015
A tumor lysate is an effective vaccine antigen for the stimulation of CD4(+) T-cell function and subsequent induction of antitumor immunity mediated by CD8(+) T cells.
Cancer biology & therapy
Kawahara M,Takaku H
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Sep 12 00:00:00 EDT 2016
Adjuvants may reduce in vivo transfection levels for DNA vaccination in mice leading to reduced antigen-specific CD8+ T cell responses.
Human vaccines & immunotherapeutics
Rose AH,Hoffmann FW,Hara JH,Urschitz J,Moisyadi S,Hoffmann PR,Bertino P
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon May 16 00:00:00 EDT 2016
Toll-Like receptor 2 (TLR2) and TLR9 play opposing roles in host innate immunity against Salmonella enterica serovar Typhimurium infection.
Infection and immunity
Zhan R,Han Q,Zhang C,Tian Z,Zhang J
35-0031 was used in Flow cytometry/Cell sorting to investigate the roles of TLR2 and TLR9 in regulating innate immunity against oral infection of Salmonella enterica serovar Typhimurium in mice.
Wed Apr 01 00:00:00 EDT 2015
The Nuclear Orphan Receptor NR2F6 Is a Central Checkpoint for Cancer Immune Surveillance.
Cell reports
Hermann-Kleiter N,Klepsch V,Wallner S,Siegmund K,Klepsch S,Tuzlak S,Villunger A,Kaminski S,Pfeifhofer-Obermair C,Gruber T,Wolf D,Baier G
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2015
Metronomic cyclophosphamide eradicates large implanted GL261 gliomas by activating antitumor Cd8+ T-cell responses and immune memory.
Oncoimmunology
Wu J,Waxman DJ
35-0031 was used in Flow cytometry/Cell sorting to prove that single-agent chemotherapy can eradicate large, established tumours and induce long-term immune memory.
Wed Apr 01 00:00:00 EDT 2015
Interleukin-25 Mediates Transcriptional Control of PD-L1 via STAT3 in Multipotent Human Mesenchymal Stromal Cells (hMSCs) to Suppress Th17 Responses.
Stem cell reports
Wang WB,Yen ML,Liu KJ,Hsu PJ,Lin MH,Chen PM,Sudhir PR,Chen CH,Chen CH,Sytwu HK,Yen BL
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Sep 08 00:00:00 EDT 2015
A Multiple Antigenic Peptide Mimicking Peptidoglycan Induced T Cell Responses to Protect Mice from Systemic Infection with Staphylococcus aureus.
PloS one
Wang XY,Huang ZX,Chen YG,Lu X,Zhu P,Wen K,Fu N,Liu BY
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue May 24 00:00:00 EDT 2016
Antibiotics in neonatal life increase murine susceptibility to experimental psoriasis.
Nature communications
Zanvit P,Konkel JE,Jiao X,Kasagi S,Zhang D,Wu R,Chia C,Ajami NJ,Smith DP,Petrosino JF,Abbatiello B,Nakatsukasa H,Chen Q,Belkaid Y,Chen ZJ,Chen W
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2015
Overexpression of IL-15 promotes tumor destruction via NK1.1+ cells in a spontaneous breast cancer model.
BMC cancer
Gillgrass AE,Chew MV,Krneta T,Ashkar AA
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Thu Apr 16 00:00:00 EDT 2015
Myeloid Sirtuin 2 Expression Does Not Impact Long-Term Mycobacterium tuberculosis Control.
PloS one
Cardoso F,Castro F,Moreira-Teixeira L,Sousa J,Torrado E,Silvestre R,Castro AG,Saraiva M,Pais TF
35-0031 was used in Flow cytometry/Cell sorting to demonstrate that Sirtuin 2 expression has a transitory effect in M. tuberculosis infection.
Tue Apr 19 00:00:00 EDT 2016
IFN-γ modulates Ly-49 receptors on NK cells in IFN-γ-induced pregnancy failure.
Scientific reports
Li ZY,Song ZH,Meng CY,Yang DD,Yang Y,Peng JP
35-0031 was used in Flow cytometry/Cell sorting to investigate the mechanism by which IFN-γ modulates Ly-49 receptors on natural killer cells during pregnancy failure.
Fri Dec 11 00:00:00 EST 2015
Rhinovirus-induced IL-25 in asthma exacerbation drives type 2 immunity and allergic pulmonary inflammation.
Science translational medicine
Beale J,Jayaraman A,Jackson DJ,Macintyre JDR,Edwards MR,Walton RP,Zhu J,Man Ching Y,Shamji B,Edwards M,Westwick J,Cousins DJ,Yi Hwang Y,McKenzie A,Johnston SL,Bartlett NW
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Oct 01 00:00:00 EDT 2014
Microbiota modulate tumoral immune surveillance in lung through a γδT17 immune cell-dependent mechanism.
Cancer research
Cheng M,Qian L,Shen G,Bian G,Xu T,Xu W,Shen G,Hu S
35-0031 was used in Flow cytometry/Cell sorting to investigate the mechanism by which commensal bacteria modulate the efficiency of tumoral immune surveillance in mucosal tissues.
Fri Aug 01 00:00:00 EDT 2014
Dietary gluten increases natural killer cell cytotoxicity and cytokine secretion.
European journal of immunology
Larsen J,Dall M,Antvorskov JC,Weile C,Engkilde K,Josefsen K,Buschard K
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Oct 01 00:00:00 EDT 2014
Orphan chemoattractant receptor GPR15 mediates dendritic epidermal T-cell recruitment to the skin.
European journal of immunology
Lahl K,Sweere J,Pan J,Butcher E
35-0031 was used in Flow cytometry/Cell sorting to investigate the role of GRP15 in dendritic epidermal T cell recruitment to the skin.
Mon Sep 01 00:00:00 EDT 2014
Viral MHC class I-like molecule allows evasion of NK cell effector responses in vivo.
Journal of immunology (Baltimore, Md. : 1950)
Pyzik M,Dumaine A,Charbonneau B,Fodil-Cornu N,Jonjic S,Vidal SM
35-0031 was used in Flow cytometry/Cell sorting to show that recognition of the viral protein m157 by the inhibitory Ly49C receptor allows escape from natural killer cell control of viral replication.
Mon Dec 15 00:00:00 EST 2014
Central role of conventional dendritic cells in regulation of bone marrow release and survival of neutrophils.
Journal of immunology (Baltimore, Md. : 1950)
Jiao J,Dragomir AC,Kocabayoglu P,Rahman AH,Chow A,Hashimoto D,Leboeuf M,Kraus T,Moran T,Carrasco-Avino G,Friedman SL,Merad M,Aloman C
35-0031 was used in Flow cytometry/Cell sorting to investigate the role of dendritic cells in neutrophil homeostasis, showing a central role in regulation of bone marrow release and survival of neutrophils.
Tue Apr 01 00:00:00 EDT 2014
Role of IL-17 and TGF-β in peritoneal adhesion formation after surgical trauma.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society
Wang G,Wu K,Li W,Zhao E,Shi L,Wang J,Shuai X,Cai K,Lu X,Tao K,Wang G
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Jun 30 00:00:00 EDT 2015
Tim-3 negatively mediates natural killer cell function in LPS-induced endotoxic shock.
PloS one
Hou H,Liu W,Wu S,Lu Y,Peng J,Zhu Y,Lu Y,Wang F,Sun Z
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Jul 06 00:00:00 EDT 2015
The nucleotide-binding leucine-rich repeat (NLR) family member NLRX1 mediates protection against experimental autoimmune encephalomyelitis and represses macrophage/microglia-induced inflammation.
The Journal of biological chemistry
Eitas TK,Chou WC,Wen H,Gris D,Robbins GR,Brickey J,Oyama Y,Ting JP
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Feb 14 00:00:00 EST 2014
Conditional deletion of ferritin h in mice reduces B and T lymphocyte populations.
PloS one
Vanoaica L,Richman L,Jaworski M,Darshan D,Luther SA,Kühn LC
35-0031 was used in Flow cytometry/Cell sorting to investigate the effect of ferritin H deletion on lymphocytes, showing that it reduces B and T lymphocyte populations.
Tue Jan 13 00:00:00 EST 2015
Prevention or early cure of type 1 diabetes by intranasal administration of gliadin in NOD mice.
PloS one
Funda DP,Fundova P,Hansen AK,Buschard K
35-0031 was used in Flow cytometry/Cell sorting to investigate whether intranasal (i.n.) administration of gliadin or gluten may arrest the diabetogenic process, showing that i.n. administration of gliadin prevents type 1 diabetes in NOD mice.
Tue Jan 13 00:00:00 EST 2015
Medullary thymic epithelial cell depletion leads to autoimmune hepatitis.
The Journal of clinical investigation
Bonito AJ,Aloman C,Fiel MI,Danzl NM,Cha S,Weinstein EG,Jeong S,Choi Y,Walsh MC,Alexandropoulos K
35-0031 was used in Flow cytometry/Cell sorting to investigate the role of TRAF6 and medullary thymic epithelial cells (mETCs) in autoimmunity, showing that mETC depletion leads to autoimmune hepatitis.
Thu Aug 01 00:00:00 EDT 2013
Induction of broad cytotoxic T cells by protective DNA vaccination against Marburg and Ebola.
Molecular therapy : the journal of the American Society of Gene Therapy
Shedlock DJ,Aviles J,Talbott KT,Wong G,Wu SJ,Villarreal DO,Myles DJ,Croyle MA,Yan J,Kobinger GP,Weiner DB
35-0031 was used in Flow cytometry/Cell sorting to investigate the efficacy of cytotoxic T cell induction by a novel polyvalent-filovirus DNA vaccine against Marburg and Ebola.
Mon Jul 01 00:00:00 EDT 2013
Lipidoid Nanoparticles Containing PD-L1 siRNA Delivered In Vivo Enter Kupffer Cells and Enhance NK and CD8(+) T Cell-mediated Hepatic Antiviral Immunity.
Molecular therapy. Nucleic acids
Dolina JS,Sung SS,Novobrantseva TI,Nguyen TM,Hahn YS
35-0031 was used in Flow cytometry/Cell sorting to demonstrate that PD-L1 knockdown on KCs is central in determining the outcome of liver viral infections, and they represent a new class of gene therapy.
Tue Feb 19 00:00:00 EST 2013
Role of NK cells in host defense against pulmonary type A Francisella tularensis infection.
Microbes and infection
Schmitt DM,O'Dee DM,Brown MJ,Horzempa J,Russo BC,Morel PA,Nau GJ
35-0031 was used in Flow cytometry/Cell sorting to investigate role of NK cells against F.tularensis infection in vivo, with LVS-vaccinated murine hosts.
Fri Mar 01 00:00:00 EST 2013
Genome-wide mouse mutagenesis reveals CD45-mediated T cell function as critical in protective immunity to HSV-1.
PLoS pathogens
Caignard G,Leiva-Torres GA,Leney-Greene M,Charbonneau B,Dumaine A,Fodil-Cornu N,Pyzik M,Cingolani P,Schwartzentruber J,Dupaul-Chicoine J,Guo H,Saleh M,Veillette A,Lathrop M,Blanchette M,Majewski J,Pearson A,Vidal SM
35-0031 was used in Flow cytometry/Cell sorting to identify host factors involved in HSV-1 and herpes simplex encephalitis susceptibility in an in vivo HSV-1 infectious model.
Sun Sep 01 00:00:00 EDT 2013
Cbx4 regulates the proliferation of thymic epithelial cells and thymus function.
Development (Cambridge, England)
Liu B,Liu YF,Du YR,Mardaryev AN,Yang W,Chen H,Xu ZM,Xu CQ,Zhang XR,Botchkarev VA,Zhang Y,Xu GL
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
Tracking antigen-specific CD4+ T cells throughout the course of chronic Leishmania major infection in resistant mice.
European journal of immunology
Pagán AJ,Peters NC,Debrabant A,Ribeiro-Gomes F,Pepper M,Karp CL,Jenkins MK,Sacks DL
35-0031 was used in Flow cytometry/Cell sorting to study parasite-specific primary CD4(+) T-cell responses to L. major in mice resistant to infection.
Fri Feb 01 00:00:00 EST 2013
CD22 is required for protection against West Nile virus Infection.
Journal of virology
Ma DY,Suthar MS,Kasahara S,Gale M,Clark EA
35-0031 was used in Flow cytometry/Cell sorting to elucidate the immunological requirements for an antibody response following West Nile virus infection.
Fri Mar 01 00:00:00 EST 2013
High frequencies of activated B cells and T follicular helper cells are correlated with disease activity in patients with new-onset rheumatoid arthritis.
Clinical and experimental immunology
Wang J,Shan Y,Jiang Z,Feng J,Li C,Ma L,Jiang Y
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2013
Markers of nonselective and specific NK cell activation.
Journal of immunology (Baltimore, Md. : 1950)
Fogel LA,Sun MM,Geurs TL,Carayannopoulos LN,French AR
35-0031 was used in Flow cytometry/Cell sorting to demonstrate that a combination of Sca-1, CD27, and KLRG1 can distinguish NK cells based on their mechanism of activation.
Sat Jun 15 00:00:00 EDT 2013
Myxomaviral Anti-Inflammatory Serpin Reduces Myeloid-Derived Suppressor Cells and Human Pancreatic Cancer Cell Growth in Mice.
Journal of cancer science & therapy
Zheng D,Chen H,Bartee MY,Williams J,Davids JA,Lomas DA,McFadden G,Lucas AR
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Aug 19 00:00:00 EDT 2013
Pulmonary T cell activation in response to chronic particulate air pollution.
American journal of physiology. Lung cellular and molecular physiology
Deiuliis JA,Kampfrath T,Zhong J,Oghumu S,Maiseyeu A,Chen LC,Sun Q,Satoskar AR,Rajagopalan S
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Feb 15 00:00:00 EST 2012
Mannose-capped lipoarabinomannan from Mycobacterium tuberculosis preferentially inhibits sphingosine-1-phosphate-induced migration of Th1 cells.
Journal of immunology (Baltimore, Md. : 1950)
Richmond JM,Lee J,Green DS,Kornfeld H,Cruikshank WW
35-0031 was used in Flow cytometry/Cell sorting to examine the ability of ManLAM to desensitize T cells to other chemoattractants during mycobacterial infection.
Sat Dec 15 00:00:00 EST 2012
CD28 promotes CD4+ T cell clonal expansion during infection independently of its YMNM and PYAP motifs.
Journal of immunology (Baltimore, Md. : 1950)
Pagán AJ,Pepper M,Chu HH,Green JM,Jenkins MK
35-0031 was used in Flow cytometry/Cell sorting to track polyclonal peptide:MHCII-specific CD4+ T cells in vivo, in mice with mutations in two sites within the cytoplasmic tail of CD28.
Sat Sep 15 00:00:00 EDT 2012
Spontaneous autoimmune gastritis and hypochlorhydria are manifest in the ileitis-prone SAMP1/YitFcs mice.
American journal of physiology. Gastrointestinal and liver physiology
Ernst PB,Erickson LD,Loo WM,Scott KG,Wiznerowicz EB,Brown CC,Torres-Velez FJ,Alam MS,Black SG,McDuffie M,Feldman SH,Wallace JL,McKnight GW,Padol IT,Hunt RH,Tung KS
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Sun Jan 01 00:00:00 EST 2012
Vaccination with synthetic constructs expressing cytomegalovirus immunogens is highly T cell immunogenic in mice.
Human vaccines & immunotherapeutics
Shedlock DJ,Talbott KT,Wu SJ,Wilson CM,Muthumani K,Boyer JD,Sardesai NY,Awasthi S,Weiner DB
35-0031 was used in Flow cytometry/Cell sorting to design a novel panel of highly-optimized synthetic vaccines encoding major CMV proteins and evaluate their immune potential.
Thu Nov 01 00:00:00 EDT 2012
Evaluation of Lassa virus vaccine immunogenicity in a CBA/J-ML29 mouse model.
Vaccine
Goicochea MA,Zapata JC,Bryant J,Davis H,Salvato MS,Lukashevich IS
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Feb 14 00:00:00 EST 2012
Species-specific activity of glycolipid ligands for invariant NKT cells.
Chembiochem : a European journal of chemical biology
Dangerfield EM,Cheng JM,Knight DA,Weinkove R,Dunbar PR,Hermans IF,Timmer MS,Stocker BL
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Jun 18 00:00:00 EDT 2012
Loss of the methyl lysine effector protein PHF20 impacts the expression of genes regulated by the lysine acetyltransferase MOF.
The Journal of biological chemistry
Badeaux AI,Yang Y,Cardenas K,Vemulapalli V,Chen K,Kusewitt D,Richie E,Li W,Bedford MT
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Jan 02 00:00:00 EST 2012
Soluble MOG35-55/I-A(b) dimers ameliorate experimental autoimmune encephalomyelitis by reducing encephalitogenic T cells.
PloS one
Gong Y,Wang Z,Liang Z,Duan H,Ouyang L,Yu Q,Xu Z,Shen G,Weng X,Wu X
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Jul 19 00:00:00 EDT 2013
Sertoli cell-specific deletion of the androgen receptor compromises testicular immune privilege in mice.
Biology of reproduction
Meng J,Greenlee AR,Taub CJ,Braun RE
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2011
T-cell recruitment and Th1 polarization in adipose tissue during diet-induced obesity in C57BL/6 mice.
Obesity (Silver Spring, Md.)
Strissel KJ,DeFuria J,Shaul ME,Bennett G,Greenberg AS,Obin MS
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2010
Expression of the chemokine binding protein M3 promotes marked changes in the accumulation of specific leukocytes subsets within the intestine.
Gastroenterology
Shang L,Thirunarayanan N,Viejo-Borbolla A,Martin AP,Bogunovic M,Marchesi F,Unkeless JC,Ho Y,Furtado GC,Alcami A,Merad M,Mayer L,Lira SA
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2009
Tracking epitope-specific T cells.
Nature protocols
Moon JJ,Chu HH,Hataye J,Pagán AJ,Pepper M,McLachlan JB,Zell T,Jenkins MK
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Tue Jun 30 00:00:00 EDT 2009
p27 deficiency cooperates with Bcl-2 but not Bax to promote T-cell lymphoma.
PloS one
Cheng N,van de Wetering CI,Knudson CM
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Wed Apr 02 00:00:00 EDT 2008
Deficient major histocompatibility complex-linked innate murine cytomegalovirus immunity in MA/My.L-H2b mice and viral downregulation of H-2k class I proteins.
Journal of virology
Xie X,Dighe A,Clark P,Sabastian P,Buss S,Brown MG
35-0031 was used in Flow cytometry/Cell sorting to show that replacement of MA/My H-2k with C57L H-2b susceptibility genes led to a remarkable loss of innate virus immunity.
Mon Jan 01 00:00:00 EST 2007
The mouse mammary carcinoma 4T1: characterization of the cellular landscape of primary tumours and metastatic tumour foci.
International journal of experimental pathology
DuPré SA,Redelman D,Hunter KW
Published figure using CD3e monoclonal antibody (Product # 35-0031-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2007
HSP70 vaccine in combination with gene therapy with plasmid DNA encoding sPD-1 overcomes immune resistance and suppresses the progression of pulmonary metastatic melanoma.
International journal of cancer
Geng H,Zhang GM,Xiao H,Yuan Y,Li D,Zhang H,Qiu H,He YF,Feng ZH
Thu Jun 01 00:00:00 EDT 2006
Functional analysis of B and T lymphocyte attenuator engagement on CD4+ and CD8+ T cells.
Journal of immunology (Baltimore, Md. : 1950)
Krieg C,Han P,Stone R,Goularte OD,Kaye J
Tue Nov 15 00:00:00 EST 2005
CCR5-deficient mice develop experimental autoimmune uveoretinitis in the context of a deviant effector response.
Investigative ophthalmology & visual science
Takeuchi A,Usui Y,Takeuchi M,Hattori T,Kezuka T,Suzuki J,Okunuki Y,Iwasaki T,Haino M,Matsushima K,Usui M
Sat Oct 01 00:00:00 EDT 2005